Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Related Categories
6 Cu Ft Fridge
Marvel Refrigerators/Freezers
Marvel Undercounter Refrigerators and Freezers
Marvel Refrigerator and Freezer Combination
Hazardous Location
Marvel Cooling Equipment
Temperature Heat Controller
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
6 Cu Ft Refrigerator
Image
Product Detail
SKU
ADA Compliant General Purpose Undercounter Refrigerator, 6 cu ft (6CAD)
Category: Refrigerator and Freezer Combination
Description: Marvel Refrigerators & Freezers
Brand:
Marvel
Model Number:
6CAD
YO-44765-10
More Details
General Purpose Undercounter Refrigerator/Freezer, 6 cu ft (6CRF)
Category: Refrigerator and Freezer Combination
Description: Marvel Refrigerators & Freezers
Brand:
Marvel
Model Number:
6CRF
YO-44765-45
More Details
Flammable Material Undercounter Refrigerator, 6 cu ft (6FAR)
Category: Refrigerator and Freezer Combination
Description: Marvel Refrigerators & Freezers
Brand:
Marvel
Model Number:
6FAR
YO-44765-55
More Details
Flammable Material Undercounter Refrigerator/Freezer, 6 cu ft (6FRF)
Category: Refrigerator and Freezer Combination
Description: Marvel Refrigerators & Freezers
Brand:
Marvel
Model Number:
6FRF
YO-44765-65
More Details
Hazardous Material Undercounter Refrigerator, 6 cu ft (6EAR)
Category: Refrigerator and Freezer Combination
Description: Marvel Refrigerators & Freezers
Brand:
Marvel
Model Number:
6EAR
YO-44765-70
More Details
Hazardous Material Undercounter Refrigerator/Freezer, 6 cu ft (6ERF)
Category: Refrigerator and Freezer Combination
Description: Marvel Refrigerators & Freezers
Brand:
Marvel
Model Number:
6ERF
YO-44765-75
More Details
General Purpose Undercounter Freezer, 6 cu ft (6CAF)
Category: Refrigerator and Freezer Combination
Description: Marvel Refrigerators & Freezers
Brand:
Marvel
Model Number:
6CAF
YO-44765-85
More Details
Laboratory undercounter refrigerator (single door); 5.4 CU. FT; 115V (REL-404A)
Category: Thermo Scientific Revco Laboratory Refrigerators
Description: Thermo Scientific Revco Undercounter Laboratory Refrigerators
Brand:
Thermo Scientific Revco
Model Number:
REL-404A
YO-44185-02
More Details
Laboratory space-saver refrigerator (single door); 11.5 CU FT; 115V (REL-1204A-20)
Category: Thermo Scientific Revco Laboratory Refrigerators
Description: Thermo Scientific Revco Laboratory Space-Saver Refrigerator with Single Door
Brand:
Thermo Scientific Revco
Model Number:
REL-1204A-20
YO-44185-10
More Details
Laboratory space-saver refrigerator (single door); 11.5 CU FT; 200/230 (REL-1204D)
Category: Thermo Scientific Revco Laboratory Refrigerators
Description: Thermo Scientific Revco Laboratory Space-Saver Refrigerator with Single Door
Brand:
Thermo Scientific Revco
Model Number:
REL-1204D
YO-44185-15
More Details
Laboratory upright refrigerator (single door); 23.3 CU FT; 115V (REL-2304-A)
Category: Thermo Scientific Revco Laboratory Refrigerators
Description: Thermo Scientific Revco Laboratory Upright Refrigerator with Single Door
Brand:
Thermo Scientific Revco
Model Number:
REL-2304-A
YO-44185-20
More Details
Laboratory upright refrigerator (single door); 23.3 CU FT; 200/230V (REL-2304D)
Category: Thermo Scientific Revco Laboratory Refrigerators
Description: Thermo Scientific Revco Laboratory Upright Refrigerator with Single Door
Brand:
Thermo Scientific Revco
Model Number:
REL-2304D
YO-44185-25
More Details
Laboratory upright refrigerator (double doors); 51.1 CU FT; 115V (REL-5004-A)
Category: Thermo Scientific Revco Laboratory Refrigerators
Description: Thermo Scientific Revco Laboratory Upright Refrigerator with Double Doors
Brand:
Thermo Scientific Revco
Model Number:
REL-5004-A
YO-44185-30
More Details
Laboratory upright refrigerator (double doors); 51.1 CU FT; 220/230V (REL-5004D)
Category: Thermo Scientific Revco Laboratory Refrigerators
Description: Thermo Scientific Revco Laboratory Upright Refrigerator with Double Doors
Brand:
Thermo Scientific Revco
Model Number:
REL-5004D
YO-44185-35
More Details
Refrigerator, Manual Defrost 24 cu ft with Casters, 115V (NSLR241WMW/0M)
Category: Nor-Lake Scientific Refrigerators
Description: Nor-Lake General Purpose Laboratory Refrigerators
Brand:
Nor-Lake
Model Number:
NSLR241WMW/0M
YO-44173-04
More Details
Refrigerator, Manual Defrost 24 cu ft with Casters, 230V (NSLR241WMW/5M)
Category: Nor-Lake Scientific Refrigerators
Description: Nor-Lake General Purpose Laboratory Refrigerators
Brand:
Nor-Lake
Model Number:
NSLR241WMW/5M
YO-44173-06
More Details
Refrigerator, Blood Bank 24 cu ft with Casters, Glass Door, 115V/60Hz (NSBR241WWG/0)
Category: Blood Bank Refrigerators
Description: Nor-Lake Blood Bank Refrigerators
Brand:
Nor-Lake
Model Number:
NSBR241WWG/0
YO-44173-22
More Details
Refrigerator, Blood Bank 24 cu ft with Casters, Glass Door, 230V/50Hz (NSBR241WWG/5)
Category: Blood Bank Refrigerators
Description: Nor-Lake Blood Bank Refrigerators
Brand:
Nor-Lake
Model Number:
NSBR241WWG/5
YO-44173-24
More Details
Refrigerator, Blood Bank 52 cu ft with Casters, 2 Glass Doors, 115V/60 (NSBR522WWG/0)
Category: Blood Bank Refrigerators
Description: Nor-Lake Blood Bank Refrigerators
Brand:
Nor-Lake
Model Number:
NSBR522WWG/0
YO-44173-26
More Details
Refrigerator, Blood Bank 52 cu ft with Casters, 2 Glass Doors, 230V/50 (NSBR522WWG/5)
Category: Blood Bank Refrigerators
Description: Nor-Lake Blood Bank Refrigerators
Brand:
Nor-Lake
Model Number:
NSBR522WWG/5
YO-44173-28
More Details
6 Cu Ft Refrigerator
Popular items by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
|
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Previous: Nor-Lake Storage Equipment
|
Next: Refrigerator only no Freezer
>
31125329-s-content_2 31125329scontent2