Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Related Categories
Ac Dc Amp
Extech Clamp Meters
Extech Instruments
Extech Equipment
Extech Electrical
Extech 700 Series 800a Clamp Meters
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
800 Amps
Image
Product Detail
SKU
Extech EX730: AC/DC TRMS Clamp Meter, 800 Amps
Category: Extech Clamp Meters
Description: Extech 700 Series 800A Clamp Meters
Brand:
Extech
Model Number:
EX730
YO-20003-20
More Details
Extech EX710: AC Clamp Meter, 800 Amps
Category: Extech Clamp Meters
Description: Extech 700 Series 800A Clamp Meters
Brand:
Extech
Model Number:
EX710
YO-20005-36
More Details
Extech EX720: True RMS AC Clamp Meter, 800 Amps
Category: Extech Clamp Meters
Description: Extech 700 Series 800A Clamp Meters
Brand:
Extech
Model Number:
EX720
YO-20005-37
More Details
Sight Flow Indicator, 0.5 To 6.5 GPM; PSF Body With 1/2" NPT(F) Connec (SFI-800-1/2-LF)
Category: Flow Indicators
Description: Sight Flow Indicators and Transmitters
Brand:
Cole-Parmer
Model Number:
SFI-800-1/2-LF
YO-32779-40
More Details
Sight Flow Indicator, 3 To 35 GPM; PSF Body With 3/4" NPT(F) Connectio (SFI-800-3/4)
Category: Flow Indicators
Description: Sight Flow Indicators and Transmitters
Brand:
Cole-Parmer
Model Number:
SFI-800-3/4
YO-32779-44
More Details
Sight Flow Indicator, 0.5 To 6.5 GPM; 1 To 10 VDC Output; Polysulfone (SFI-800-1/2-LF-A712)
Category: Flow Indicators
Description: Sight Flow Indicators and Transmitters
Brand:
Cole-Parmer
Model Number:
SFI-800-1/2-LF-A712
YO-32779-54
More Details
Sight Flow Indicator, 2 To 20 GPM; 1 To 10 VDC Output; Polysulfone Bod (SFI-800-1/2-A712)
Category: Flow Indicators
Description: Sight Flow Indicators and Transmitters
Brand:
Cole-Parmer
Model Number:
SFI-800-1/2-A712
YO-32779-56
More Details
Sight Flow Indicator, 3 To 35 GPM; 1 To 10 VDC Output; Polysulfone Bod (SFI-800-3/4-A712)
Category: Flow Indicators
Description: Sight Flow Indicators and Transmitters
Brand:
Cole-Parmer
Model Number:
SFI-800-3/4-A712
YO-32779-58
More Details
PTFE tubing, 8.00 x 9 mm, 25 ft/pk (YO-06363-20)
Category: PTFE Tubing
Description: Metric PTFE tubing
Brand:
Cole-Parmer
YO-06363-20
More Details
Tip Art 100-1000µL Sterile 800/pk (2079E)
Category: MBP ART Aerosol-Resistant Pipette Tips
Description: Racked, presterilized tips
Brand:
ART
Model Number:
2079E
YO-25716-56
More Details
Tip Art 100-1000µL Sterile 800/pk (2079)
Category: MBP ART Aerosol-Resistant Pipette Tips
Description: Racked, presterilized tips
Brand:
ART
Model Number:
2079
YO-25716-58
More Details
Cole-Parmer 150-mm Direct Reading Flowmeter,Air,Ss/Ss Float; 800 mL/m (YO-32032-09)
Category: Cole-Parmer Direct Reading Panel-Mount Flowmeters
Description: Cole-Parmer 150-mm Direct Reading Panel-Mount Flowmeters
Brand:
Cole-Parmer
YO-32032-09
More Details
Cole-Parmer 150-mm Direct Reading Flowmeter,Air,Ss/Ss Float; 800 mL/m (YO-32035-09)
Category: Cole-Parmer Direct Reading Panel-Mount Flowmeters
Description: Cole-Parmer 150-mm Direct Reading Panel-Mount Flowmeters
Brand:
Cole-Parmer
YO-32035-09
More Details
Panel-mount PTFE/PFA correlated flowmeter, 3/8" NPT(F), 800 to 2,500 m (YO-32050-12)
Category: Variable Area Flowmeters
Description: Cole-Parmer Heavy-Duty PFA Correlated Flowmeters
Brand:
Cole-Parmer
YO-32050-12
More Details
Full-PTFE/PFA in-line flowmeter, 3/8" NPT(F), 800 to 2,500 ml/min (YO-32056-12)
Category: Variable Area Flowmeters
Description: Cole-Parmer Safe-Design PFA Flowmeters
Brand:
Cole-Parmer
YO-32056-12
More Details
Full-PTFE/PFA in-line flowmeter, 3/8" NPT(F), with valve, 800 to 2,500 (YO-32057-12)
Category: Variable Area Flowmeters
Description: Cole-Parmer Safe-Design PFA Flowmeters
Brand:
Cole-Parmer
YO-32057-12
More Details
Flowmeter, In-line, With Brass Fittings; 800 SLPM O2 (YO-32605-25)
Category: Variable Area Flowmeters
Description: Cole-Parmer Direct-Reading Multigas Flowmeters
Brand:
Cole-Parmer
YO-32605-25
More Details
Flowmeter, In-line, With Stainless Steel Fittings; 800 SLPM O2/s (YO-32605-30)
Category: Variable Area Flowmeters
Description: Cole-Parmer Direct-Reading Multigas Flowmeters
Brand:
Cole-Parmer
YO-32605-30
More Details
Flowmeter, In-line With Valve, With Brass Fittings; 800 SLPM O2/ (YO-32605-35)
Category: Variable Area Flowmeters
Description: Cole-Parmer Direct-Reading Multigas Flowmeters
Brand:
Cole-Parmer
YO-32605-35
More Details
Flowmeter, In-line With Valve, With Stainless Steel Fittings; 800 SLPM (YO-32605-40)
Category: Variable Area Flowmeters
Description: Cole-Parmer Direct-Reading Multigas Flowmeters
Brand:
Cole-Parmer
YO-32605-40
More Details
800 Amps
Popular items by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
|
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Previous: Brass Air Line Fittings
|
Next: 2.5 X3 0 Brass Fitting
>
31125329-s-content_2 31125329scontent2