Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Genesys 10 Uv Scanning Spectrophotometer
Also showing items for Genesys 10 Uv, Genesys 10 Spectrophotometer and Genesys 10 Uv Spectrophotometer
Image
Product Detail
SKU
Thermo Scientific GENESYS™ 10S UV-Visible Spectrophotometer USB; Si
Category: Visible/UV-Visible Spectroscopy
Description: Thermo Scientific GENESYS 10S Visible and UV/Visible Scanning Spectrophotometers
Brand:
Thermo Scientific Genesys
YO-02654-20
More Details
Thermo Scientific GENESYS™ 10S UV-Visible Spectrophotometer USB wit
Category: Visible/UV-Visible Spectroscopy
Description: Thermo Scientific GENESYS 10S Visible and UV/Visible Scanning Spectrophotometers
Brand:
Thermo Scientific Genesys
YO-02654-22
More Details
Peltier thermostated single cell holder for GENESYS 10 (222-238100)
Category: Visible/UV-Visible Spectroscopy
Description: Thermo Scientific GENESYS 10 Scanning UV/Visible Spectrophotometer
Brand:
Thermo Scientific Genesys
Model Number:
222-238100
YO-02653-90
More Details
GENESYS 10 nanoCell accessory, 0.2mm pathlength (222-227700)
Category: Visible/UV-Visible Spectroscopy
Description: Thermo Scientific GENESYS 10 Scanning UV/Visible Spectrophotometer
Brand:
Thermo Scientific Genesys
Model Number:
222-227700
YO-02653-95
More Details
GENESYS 10 nanoCell accessory, 1.0mm pathlength (222-227800)
Category: Visible/UV-Visible Spectroscopy
Description: Thermo Scientific GENESYS 10 Scanning UV/Visible Spectrophotometer
Brand:
Thermo Scientific Genesys
Model Number:
222-227800
YO-02653-96
More Details
GENESYS 10 nanoCell accessory, 0.2mm pathlength adapter (222-227900)
Category: Visible/UV-Visible Spectroscopy
Description: Thermo Scientific GENESYS 10 Scanning UV/Visible Spectrophotometer
Brand:
Thermo Scientific Genesys
Model Number:
222-227900
YO-02653-97
More Details
GENESYS 10 nanoCell accessory, 1.0mm pathlength adapter (222-228000)
Category: Visible/UV-Visible Spectroscopy
Description: Thermo Scientific GENESYS 10 Scanning UV/Visible Spectrophotometer
Brand:
Thermo Scientific Genesys
Model Number:
222-228000
YO-02653-98
More Details
GENESYS 10 nanoCell accessory with 0.2mm and 1.0mm pathlength adapter (222-230300)
Category: Visible/UV-Visible Spectroscopy
Description: Thermo Scientific GENESYS 10 Scanning UV/Visible Spectrophotometer
Brand:
Thermo Scientific Genesys
Model Number:
222-230300
YO-02653-99
More Details
Thermo Scientific GENESYS™ 10S Bio UV/Visible Spectrophotometer, 6
Category: Visible/UV-Visible Spectroscopy
Description: Thermo Scientific GENESYS 10S Bio UV/Visible Scanning Spectrophotometers
Brand:
Thermo Scientific Genesys
YO-02654-00
More Details
Thermo Scientific GENESYS™ 10S Bio UV/Visible Spectrophotometer wit
Category: Visible/UV-Visible Spectroscopy
Description: Thermo Scientific GENESYS 10S Bio UV/Visible Scanning Spectrophotometers
Brand:
Thermo Scientific Genesys
YO-02654-02
More Details
Thermo Scientific GENESYS™ 10S Visible Spectrophotometer USB; Singl
Category: Visible/UV-Visible Spectroscopy
Description: Thermo Scientific GENESYS 10S Visible and UV/Visible Scanning Spectrophotometers
Brand:
Thermo Scientific Genesys
YO-02654-10
More Details
Thermo Scientific GENESYS™ 10S Visible Spectrophotometer USB with P
Category: Visible/UV-Visible Spectroscopy
Description: Thermo Scientific GENESYS 10S Visible and UV/Visible Scanning Spectrophotometers
Brand:
Thermo Scientific Genesys
YO-02654-12
More Details
Genesys 10 Uv Scanning Spectrophotometer
Popular items by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
|
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Previous: Genesys Visible/Uv-Visible Spectroscopy
|
Next: Thermo Genesys 10
>
31125329-s-content_2 31125329scontent2