Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Related Categories
Id 1 4 Od Rubber Tubing
10 Od Stainless Tubing
2.065 Od Stainless Steel Tubing
875 Inch Od Stainless Steel Tubing
8 in Od Stainless Steel Tube
Thin Wall Stainless Tubing 1.5 Od
Thin Wall Stainless Tubing 11 Id
Food Grade Stainless Tubing
3 Mm Diameter Stainless Steel Tube
8 Inch Od Stainless Steel Tube
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, Fittings, and Hose
Id 3 16 Od Stainless Tubing
Image
Product Detail
SKU
Ester-Based Polyurethane Tubing; 1/8" ID, 3/16" OD (YO-95680-02)
Category: Polyurethane tubing
Description: Ester-Based Polyurethane Tubing
Brand:
Cole-Parmer
YO-95680-02
More Details
Tygon® high-purity tubing, 1/16"ID x 3/16"OD, 50 ft/pack (YO-95635-00)
Category: Tubing for Bioprocessing
Description: TYGON High-Purity Tubing
Brand:
Tygon
YO-95635-00
More Details
ReSeal Sanitary Coupling, 3/8" ID, 11/16" OD, 3/8" Clamp
Category: Sanitary Fittings, Clamps, and Gaskets
Description: ReSeal Sanitary Couplings
Brand:
ReSeal
YO-31511-08
More Details
ReSeal Sanitary Coupling, 1/2" ID, 13/16" OD, 1/2" Clamp
Category: Sanitary Fittings, Clamps, and Gaskets
Description: ReSeal Sanitary Couplings
Brand:
ReSeal
YO-31511-13
More Details
Ester-Based Polyurethane Tubing; 1/4" ID, 7/16" OD (YO-95680-16)
Category: Polyurethane tubing
Description: Ester-Based Polyurethane Tubing
Brand:
Cole-Parmer
YO-95680-16
More Details
316 Stainless Steel Tubing; dimensions, 5/16" ID, 3/8" OD, 1/32" wall; (YO-03300-15)
Category: Rigid Tubing
Description: 316 Stainless Steel Tubing
Brand:
Cole-Parmer
YO-03300-15
More Details
Tygon Lab Tubing, Non-DEHP, 1/16"ID x 3/16"OD, 50 ft/pack
Category: TYGON Lab Tubing
Description: TYGON Lab Tubing, non-DEHP
Brand:
Tygon
YO-06407-72
More Details
Tygon lab tubing, Non-DEHP, 1/16"ID x 3/16"OD, 50 ft/pack (ACF00003-CP)
Category: TYGON Lab Tubing
Description: TygonE - 3603 Lab Tubing
Brand:
Tygon
Model Number:
ACF00003-CP
YO-07407-72
More Details
Tygon 2375 Ultra Chemical-Resistant Tubing, 1/16"ID x 3/16"OD, 50 ft/p
Category: Tygon Tubing
Description: Tygon 2375 Ultra Chemical Resistant Tubing
Brand:
Tygon
YO-95630-11
More Details
TYGON® Food and Beverage Tubing, 1/16"ID x 3/16"OD, 50 ft/pack
Category: Tygon
®
Tubing
Description: TYGON® Food and Beverage Tubing
Brand:
Tygon
YO-95631-01
More Details
Ester-Based Polyurethane Tubing; 3/16" ID, 5/16" OD (YO-95680-08)
Category: Polyurethane tubing
Description: Ester-Based Polyurethane Tubing
Brand:
Cole-Parmer
YO-95680-08
More Details
Tygon Silicone Tubing, 1/16"ID x 3/16"OD, 50 ft/pack
Category: Flexible Tubing
Description: Tygon Silicone Tubing
Brand:
Tygon
YO-95702-02
More Details
Norprene food tubing, 1/16"ID x 3/16"OD, 50 ft/pack
Category: Flexible Tubing
Description: Norprene Food-Grade Tubing
Brand:
Norprene
YO-95710-00
More Details
Tygon ultra chemical-resistant tubing, 1/16"ID x 3/16"OD, 50 ft/pack
Category: Tygon Tubing
Description: Tygon 2375 Ultra Chemical Resistant Tubing
Brand:
Tygon
YO-95884-61
More Details
Sanipure Tubing, 1/16" ID x 3/16" OD, 50ft
Category: Flexible Tubing
Description: Sanipure Tubing
Brand:
Sanipure
YO-96132-04
More Details
Norprene Tubing, 1/16"ID x 3/16"OD, 50 ft/pack
Category: Norprene Tubing
Description: Norprene Tubing, 50 ft/pack
Brand:
Norprene
YO-06410-01
More Details
Bev-A-Line V Tubing, 1/16"ID x 3/16"OD, 50 ft/pack
Category: Rigid Tubing
Description: Bev-A-Line V Tubing
Brand:
Bev-A-Line
YO-06491-10
More Details
Tygon High Purity Tubing, 1/16"ID x 3/16"OD, 50 ft/pk
Category: Flexible Tubing
Description: Tygon High-Purity Tubing
Brand:
Tygon
YO-95635-50
More Details
Tygon Food and Beverage Tubing, non-DEHP, 1/16"ID x 3/16"OD, 50 ft/pac
Category: Tygon Tubing
Description: TYGON Food and Beverage Tubing, non-DEHP
Brand:
Tygon
YO-95632-01
More Details
Tygon fuel and lubricant tubing, 3/32"ID x 3/16"OD, 50 ft/pack
Category: Tygon Tubing
Description: TYGON Fuel and Lubricant Tubing
Brand:
Tygon
YO-95633-00
More Details
Id 3 16 Od Stainless Tubing
Popular items by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
|
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Previous: 12 in Flexible Tubing
|
Next: 2 Od 16 Ga Ss Tubing
>
31125329-s-content_2 31125329scontent2