Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 100
Thermocouple Type B 18
to
Oakton Magnetic
Image
Product Detail
More Detail
Thermocouple Type B 18
Buy Thermocouple Type B 18 Items Including:
Type T Fine-gauge PTFE-insulated thermocouple probe (IT-18)
Vacuum Research Thermocouple Sensor, 1/8" NPT (m).
More Details
Thermocouple 24"
Buy Thermocouple 24" Items Including:
Uniwell Universal Industrial Thermocouple Probes Type J, 24" Length (1910-J-24-1/2-W2-446)
Type T PVC-insulated thermocouple probe with epoxy-coated tip; 24-gaug (113001I-T)
Type K PTFE moving surface thermocouple probe; 2.4"L (112801I-K).
More Details
Velcro Temperature Thermocouple Instruments and Probes
Buy Velcro Temperature Thermocouple Instruments and Probes Items Including:
Type J Velcro strap-on thermocouple probe; for 0.75 - 2.75 OD
Type K Velcro strap-on thermocouple probe; for 0.75 - 2.75 OD.
More Details
Velcro Strap-on Surface Probes
Buy Velcro Strap-on Surface Probes Items Including:
Type J Velcro strap-on thermocouple probe; for 0.75 - 2.75 OD
Type K Velcro strap-on thermocouple probe; for 0.75 - 2.75 OD.
More Details
Velcro Thermocouple Instruments and Probes
Buy Velcro Thermocouple Instruments and Probes Items Including:
Type J Velcro strap-on thermocouple probe; for 0.75 - 2.75 OD
Type K Velcro strap-on thermocouple probe; for 0.75 - 2.75 OD.
More Details
Velcro Oakton Surface Probes
Buy Velcro Oakton Surface Probes Items Including:
Type J Velcro strap-on thermocouple probe; for 0.75 - 2.75 OD
Type K Velcro strap-on thermocouple probe; for 0.75 - 2.75 OD.
More Details
Velcro Thermocouple Probes
Buy Velcro Thermocouple Probes Items Including:
Type J Velcro strap-on thermocouple probe; for 0.75 - 2.75 OD
Type K Velcro strap-on thermocouple probe; for 0.75 - 2.75 OD.
More Details
Velcro Temperature Products
Buy Velcro Temperature Products Items Including:
Type J Velcro strap-on thermocouple probe; for 0.75 - 2.75 OD
Type K Velcro strap-on thermocouple probe; for 0.75 - 2.75 OD.
More Details
Velcro Temperature, Humidity and Moisture Products
Buy Velcro Temperature, Humidity and Moisture Products Items Including:
Type J Velcro strap-on thermocouple probe; for 0.75 - 2.75 OD
Type K Velcro strap-on thermocouple probe; for 0.75 - 2.75 OD.
More Details
Velcro Strap
Buy Velcro Strap Items Including:
Type J Velcro strap-on thermocouple probe; for 0.75 - 2.75 OD
Type K Velcro strap-on thermocouple probe; for 0.75 - 2.75 OD.
More Details
Oakton Probes
Buy Oakton Probes Items Including:
Oakton ATC probe for pH 600, pH 610, pH 620, PC 650, PD 650, PCD 650 m (YO-35418-05)
Oakton ATC probe for pH 600, pH 610, pH 620, PC 650, PD 650, PCD 650 m (YO-35418-07)
Wd-08500-65:T-Pentrtn Probe Type T Thermocouple Penetration Probe (052297A-T).
More Details
Oakton Temperature Probes
Buy Oakton Temperature Probes Items Including:
Oakton ATC probe for pH 600, pH 610, pH 620, PC 650, PD 650, PCD 650 m (YO-35418-05)
Oakton ATC probe for pH 600, pH 610, pH 620, PC 650, PD 650, PCD 650 m (YO-35418-07)
Type T Pipe-fitting thermocouple probes; 12"L immersion depth (072997D-T).
More Details
Type K Thermocouple Instruments
Buy Type K Thermocouple Instruments Items Including:
Oakton Temp-10 type J thermocouple thermometer with boot
OAKTON Temp-10 type K thermocouple thermometer with boot
Oakton Temp-10 type T thermocouple thermometer with boot
Oakton Temp-10 type J thermocouple thermometer with boot and NIST-trac.
More Details
Oakton Surface Probes - Standard
Buy Oakton Surface Probes - Standard Items Including:
Wd-08500-60:T-Surface Probe Type T Thermocouple Surface Probe (090697C-T)
Type K Standard Surface Thermocouple Probe; 10"L (090697C-K).
More Details
Oakton Surface Probes - Small-Diameter
Buy Oakton Surface Probes - Small-Diameter Items Including:
Type T Small-diameter surface thermocouple probe; 8"L (062097C-J)
Type K Small-diameter surface thermocouple probe; 8"L (062097C-K).
More Details
Surface Probes - Small-Diameter
Buy Surface Probes - Small-Diameter Items Including:
Type T Small-diameter surface thermocouple probe; 8"L (062097C-J)
Type K Small-diameter surface thermocouple probe; 8"L (062097C-K).
More Details
Small do Probe
Buy Small do Probe Items Including:
Small-diameter probes with hypodermic tip, 4" L, J type (060997C-J)
700 Series Interchangeable Thermistor Small Surface Probe
400 Series Thermistor small flexible probe (402)
Small flexible probe, vinyly sheath and tip
10-ft lead
More Details
Thermocouple Adhesive
Buy Thermocouple Adhesive Items Including:
Type J Self-Adhesive Probe (031804M-J)
Type K Self-Adhesive Probe (031804M-K).
More Details
Oakton Self-Adhesive Surface Probes
Buy Oakton Self-Adhesive Surface Probes Items Including:
Type J Self-Adhesive Probe (031804M-J)
Type K Self-Adhesive Probe (031804M-K).
More Details
Self-Adhesive Surface Probes
Buy Self-Adhesive Surface Probes Items Including:
Type J Self-Adhesive Probe (031804M-J)
Type K Self-Adhesive Probe (031804M-K).
More Details
Self Adhesive Products
Buy Self Adhesive Products Items Including:
Type J Self-Adhesive Probe (031804M-J)
Type K Self-Adhesive Probe (031804M-K)
Type T Self-Adhesive Probe (031804M-T).
More Details
Moving Surface Temperature Probes
Buy Moving Surface Temperature Probes Items Including:
Type T Roller thermocouple probe; 10.5"L with 4"L bow (112801H-T)
Type E Roller thermocouple probe; 10.5"L with 4"L bow (112801H-E)
Type K Roller thermcouple probe; 10.5"L with 4"L bow (112801H-K).
More Details
Oakton Moving Surface Probes with Ptfe-Coating
Buy Oakton Moving Surface Probes with Ptfe-Coating Items Including:
Type K Rolling wheel moving surface thermocouple probe (08445-31)
Type K PTFE moving surface thermocouple probe; 2.4"L (112801I-K).
More Details
Moving Surface Probes with Ptfe-Coating
Buy Moving Surface Probes with Ptfe-Coating Items Including:
Type K Rolling wheel moving surface thermocouple probe (08445-31)
Type K PTFE moving surface thermocouple probe; 2.4"L (112801I-K).
More Details
Themocouple K Type
Buy Themocouple K Type Items Including:
Waterproof Themocouple and Thermistor Datalogger Thermometer (367DA367)
Type J PTFE moving surface themocouple probe; 2.4"L (112801I-J).
More Details
Oakton Moving Surface Probe with Roller
Buy Oakton Moving Surface Probe with Roller Items Including:
Type T Roller thermocouple probe; 10.5"L with 4"L bow (112801H-T)
Type E Roller thermocouple probe; 10.5"L with 4"L bow (112801H-E)
Type K Roller thermcouple probe; 10.5"L with 4"L bow (112801H-K).
More Details
Moving Surface Probe with Roller
Buy Moving Surface Probe with Roller Items Including:
Type T Roller thermocouple probe; 10.5"L with 4"L bow (112801H-T)
Type E Roller thermocouple probe; 10.5"L with 4"L bow (112801H-E)
Type K Roller thermcouple probe; 10.5"L with 4"L bow (112801H-K).
More Details
Magnetic Thermocouple Probe
Buy Magnetic Thermocouple Probe Items Including:
Type T Dropping/magnetic thermocouple probe; 1.5L (072197A-T)
Type K Dropping/magnetic thermocouple probe; 1.5"L (113001B-K)
Type J Dropping/magnetic thermocouple probe; (113001B-J)
Type T Dropping/magnetic thermocouple probe (113001B-T).
More Details
Magnetic Temperature Probe
Buy Magnetic Temperature Probe Items Including:
Type T Dropping/magnetic thermocouple probe; 1.5L (072197A-T)
Type J Dropping/magnetic thermocouple probe; (113001B-J)
Type T Dropping/magnetic thermocouple probe (113001B-T)
Magnetic Surface Temperature Probe Kit for Dew Point Meter (YO-37962-02).
More Details
Oakton Magnetic / Dropping Surface Probes
Buy Oakton Magnetic / Dropping Surface Probes Items Including:
Type K Dropping/magnetic thermocouple probe; 1.5"L (113001B-K)
Type K Dropping/magnetic thermocouple probe; 1.5"L (072197A-K)
Type J Dropping/magnetic thermocouple probe; 1.5"L (072197A-J).
More Details
70618
results
<<<Previous
96
97
98
99
100
101
102
103
104
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
10
20
30
40
50
60
70
80
85
86
87
88
89
90
91
92
93
94
95
96
97
98
99
100
101
102
103
104
105
106
107
108
109
110
111
112
113
114
120
130
140
150
160
170
180
190
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Thermocouple Type B 18 to Oakton Magnetic
>
31125329-s-toc_2 31125329stoc2