Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 11
Chemfluor Female Straight Couplers
to
Ptfe Bulkhead Unions
Image
Product Detail
More Detail
Chemfluor Female Straight Couplers
Buy Chemfluor Female Straight Couplers Items Including:
Pipe fittings, Straight coupling, Chemfluor® PTFE, 1/8" NPT(F), 15 (YO-06469-16).
More Details
Npt Female Straight Pipe Coupling
Buy Npt Female Straight Pipe Coupling Items Including:
Pipe fittings, Straight coupling, Chemfluor® PTFE, 1/8" NPT(F), 15 (YO-06469-16).
More Details
Pipe Fitting Couplers
Buy Pipe Fitting Couplers Items Including:
Pipe fittings, Straight coupling, Chemfluor® PTFE, 1/8" NPT(F), 15 (YO-06469-16).
More Details
Npt Pipe Couplers
Buy Npt Pipe Couplers Items Including:
Pipe fittings, Straight coupling, Chemfluor® PTFE, 1/8" NPT(F), 15 (YO-06469-16).
More Details
Tubing Couplers
Buy Tubing Couplers Items Including:
Coupling Body, Polysulfone, 3/4" ID Tube, 10/pk (MPU171239)
Cam coupler plug, 1", Food Grade Polypropylene, Each (YO-31516-67)
Male luer x female luer coupler, PP, 25/pk (YO-45500-82)
Male Cam X 3/4-in NPT(M) Adapter, PVDF (QC-F075-PVDF).
More Details
Plastic Bulkhead Fittings 2
Buy Plastic Bulkhead Fittings 2 Items Including:
Thermo Scientific Nalgene barbed bulkhead fitting kit, 1/2" tubing ID
Bulkhead Fitting, PVDF, 1/2" NPT(F) Port X 2-1/16" OD (FK-TA04)
Bulkhead Fitting, PVDF, 1-1/4" NPT(F) Port X 2-7/8" OD (FK-TA10).
More Details
Pargrip Pvdf Tube Fittings
Buy Pargrip Pvdf Tube Fittings Items Including:
Bulkhead Fitting, PVDF, 3/8" NPT(F) Port X 1-13/16" OD (FK-TA03)
Barbed to sanitary adapter, 1/4" to mini sanitary, PVDF, pack of 10 (YO-31513-80)
Barbed to sanitary adapter, 3/8" to mini sanitary, PVDF, pack of 10 (YO-31513-81).
More Details
Hydraulic Fittings 1 8 Tubing Fittings
Buy Hydraulic Fittings 1 8 Tubing Fittings Items Including:
Polypropylene Sanitary Barbed Adapter, 3/4" Clamp Size x 1/8" Tubing I (YO-31805-24)
PVDF Sanitary Barbed Adapter, 3/4" Clamp Size x 1/8" Tubing ID (YO-31808-24)
Stopcock with Compression Fittings, 1/8" tube OD (MV-14-018).
More Details
Pvdf Bulkhead Connectors
Buy Pvdf Bulkhead Connectors Items Including:
Bulkhead Fitting, PVDF, 3/8" NPT(F) Port X 1-13/16" OD (FK-TA03)
Bulkhead Fitting, PVDF, 1-1/4" NPT(F) Port X 2-7/8" OD (FK-TA10)
Push-to-connect Bulkhead Union Connector, PVDF, 1/4" Tube OD, Pack Of (F4BU4).
More Details
Cole-Parmer Pvdf Bulkhead Connectors
Buy Cole-Parmer Pvdf Bulkhead Connectors Items Including:
Bulkhead Fitting, PVDF, 3/8" NPT(F) Port X 1-13/16" OD (FK-TA03)
Bulkhead Fitting, PVDF, 1-1/4" NPT(F) Port X 2-7/8" OD (FK-TA10)
Bulkhead Fitting, PVDF, 1-1/2" NPT(F) Port X 3-1/8" OD (FK-TA12).
More Details
Cole Pvdf Bulkhead Connectors
Buy Cole Pvdf Bulkhead Connectors Items Including:
Bulkhead Fitting, PVDF, 3/8" NPT(F) Port X 1-13/16" OD (FK-TA03)
Bulkhead Fitting, PVDF, 1-1/4" NPT(F) Port X 2-7/8" OD (FK-TA10)
Bulkhead Fitting, PVDF, 1-1/2" NPT(F) Port X 3-1/8" OD (FK-TA12).
More Details
Dasko 1 2 Fitting
Buy Dasko 1 2 Fitting Items Including:
Compression To Thread Adapter, Stainless Steel, 10mm OD Tubing X 1/2" (YO-31528-30)
Push-to-connect male threaded adapter, 316 stainless steel, 1/2" x 1/2 (YO-31370-05)
Aseptiquik-X, High Flow, Body, 1-1/2" Sanitary, High Temperature.
More Details
Cole Fittings
Buy Cole Fittings Items Including:
Antifoam probe for 3- and 5-liter systems
Fits 10-mm ports; 130-mm im (YO-01919-62)
Antifoam probe for 7- and 15-liter systems
Fits 10-mm ports; 170-mm i (YO-01919-64)
Barbed fitting kits, BSPT, adapters/elbows/tees, Natural PP (YO-31514-30).
More Details
G1 2 Thread Fittings
Buy G1 2 Thread Fittings Items Including:
Compression To Thread Adapter, Stainless Steel, 10mm OD Tubing X 1/2" (YO-31528-30)
Reducing Nipple, Polypropylene, 1/2" x 1/8", 100 PACK (YO-30627-47)
Reducing Nipple, Polypropylene, 1/2" x 1/4", 100 PACK (YO-30627-51).
More Details
Ptfe Tubing, Fittings, and Hose
Buy Ptfe Tubing, Fittings, and Hose Items Including:
Assembly Of Flared 0.040 X 0.066" PTFE Tubing, 36", With 1/4-28 Fittin (YO-96154-03)
Bulkhead Fitting, PTFE, 1/4" NPT(F) Port X 1-11/16" OD (F-TA02)
Bulkhead Fitting, PTFE, 3/8" NPT(F) Port X 1-13/16" OD (F-TA03).
More Details
Ptfe Fittings
Buy Ptfe Fittings Items Including:
PTFE Fittings, Cone1/8" OD4/pack (YO-06471-62)
PTFE Fittings, Replacement O-rings, 2 to 4 mm, 10/pack (YO-06473-26)
PTFE Fittings, Grippers, 1/16" size, 10/pack (YO-06473-60)
PTFE Fittings, Grippers, 1/8" size, 10/pack (YO-06473-65).
More Details
Tubing Bulkhead Fitting
Buy Tubing Bulkhead Fitting Items Including:
Thermo Scientific Nalgene barbed bulkhead fitting kit, 1/2" tubing ID
Thermo Scientific Nalgene barbed bulkhead fitting kit, 1/4" tubing ID
Bulkhead Fitting, PTFE, 1/4" NPT(F) Port X 1-11/16" OD (F-TA02).
More Details
Nptf Fittings
Buy Nptf Fittings Items Including:
Metal Quick-Disconnect NPT(F) Pipe Adapter Insert; Valved, 1/4" NPT(F) (MCD2604)
Bulkhead Fitting, PTFE, 1/4" NPT(F) Port X 1-11/16" OD (F-TA02)
Bulkhead Fitting, PVDF, 3/8" NPT(F) Port X 1-13/16" OD (FK-TA03).
More Details
Bulkhead Connector Hose False
Buy Bulkhead Connector Hose False Items Including:
Bulkhead Fitting, PTFE, 1/4" NPT(F) Port X 1-11/16" OD (F-TA02)
Bulkhead Fitting, PTFE, 3/8" NPT(F) Port X 1-13/16" OD (F-TA03)
Bulkhead Fitting, PVDF, 3/8" NPT(F) Port X 1-13/16" OD (FK-TA03).
More Details
Threaded Tubing Connectors
Buy Threaded Tubing Connectors Items Including:
Male branch tee, polyethylene, 1/8" NPT branch x 1/4" tubing ID, 100/p (YO-30401-67)
Male branch tee, HDPE, 1/8" NPT branch x 1/4" tubing ID, 10/pack
Bulkhead Fitting, PTFE, 1/4" NPT(F) Port X 1-11/16" OD (F-TA02).
More Details
Union Threaded Fitting
Buy Union Threaded Fitting Items Including:
Union threaded elbow, PFA, 1/8" NPTF x 1/8" NPTF (YO-06546-32)
Bulkhead Fitting, PTFE, 1/4" NPT(F) Port X 1-11/16" OD (F-TA02)
Bulkhead Fitting, PTFE, 3/8" NPT(F) Port X 1-13/16" OD (F-TA03).
More Details
Ptfe Bulkhead Connectors
Buy Ptfe Bulkhead Connectors Items Including:
Bulkhead Fitting, PTFE, 1/4" NPT(F) Port X 1-11/16" OD (F-TA02)
Bulkhead Fitting, PTFE, 3/8" NPT(F) Port X 1-13/16" OD (F-TA03)
Micro Tubing Valves; 2-way, Bulkhead; Tube OD, 1 To 4 mm; 2/pack (YO-06473-13).
More Details
T Hose Connector with Thread
Buy T Hose Connector with Thread Items Including:
Male Branch Tee, 316 SS, 10-32 UNF(M) x 1/16" hose barb (YO-31209-50)
Male Branch Tee, 316 SS, 10-32 UNF(M) x 1/8" hose barb (YO-31209-51)
Bulkhead Fitting, PTFE, 1/4" NPT(F) Port X 1-11/16" OD (F-TA02).
More Details
Tubing Bulkhead
Buy Tubing Bulkhead Items Including:
Thermo Scientific Nalgene barbed bulkhead fitting kit, 1/2" tubing ID
Thermo Scientific Nalgene barbed bulkhead fitting kit, 1/4" tubing ID
High-Flow Quick-Disconnect Hose Barb; Valved, PS, 5/8" Tube ID (HFCD161035).
More Details
Hermetic Bulkhead Tube Fittings
Buy Hermetic Bulkhead Tube Fittings Items Including:
Micro Tubing Valves; 2-way, Bulkhead; Tube OD, 1 To 4 mm; 2/pack (YO-06473-13)
Bulkhead Fitting, PTFE, 1/4" NPT(F) Port X 1-11/16" OD (F-TA02)
Bulkhead Fitting, PTFE, 3/8" NPT(F) Port X 1-13/16" OD (F-TA03).
More Details
Pipe to Tubing Od Tubing Fittings
Buy Pipe to Tubing Od Tubing Fittings Items Including:
Bulkhead Fitting, PTFE, 1/4" NPT(F) Port X 1-11/16" OD (F-TA02)
Bulkhead Fitting, PTFE, 3/8" NPT(F) Port X 1-13/16" OD (F-TA03)
Bulkhead Fitting, PVDF, 3/8" NPT(F) Port X 1-13/16" OD (FK-TA03).
More Details
Npt(F) Unions
Buy Npt(F) Unions Items Including:
NPT(F) union elbows, nylon, 1/8", 6/pack (YO-06349-60)
Hayward True Union Ball Check Valve, 1" socket and NPT(F) connectors
Straight Union Coupling, PTFE, 1/8" NPT(F) (F-SC01)
Straight Union Coupling, PTFE, 3/8" NPT(F) (F-SC03).
More Details
Union Tube Fittings
Buy Union Tube Fittings Items Including:
Compression Reducing Union, Stainless Steel, 12mm X 6mm OD Tubing (YO-31528-41)
Compression Straight Union, Stainless Steel, 10mm OD Tubing (YO-31528-35)
Compression Straight Union, Stainless Steel, 12mm OD Tubing (YO-31528-36).
More Details
Ptfe Hose Fittings
Buy Ptfe Hose Fittings Items Including:
Bulkhead Fitting, PTFE, 1/4" NPT(F) Port X 1-11/16" OD (F-TA02)
Bulkhead Fitting, PTFE, 3/8" NPT(F) Port X 1-13/16" OD (F-TA03)
Assembly Of Flared 0.040 X 0.066" PTFE Tubing, 18", With 1/4-28 Fittin (YO-96154-01).
More Details
Ptfe Bulkhead Unions
Buy Ptfe Bulkhead Unions Items Including:
Micro Tubing Valves; 2-way, Bulkhead; Tube OD, 1 To 4 mm; 2/pack (YO-06473-13)
Bulkhead Fitting, PTFE, 1/4" NPT(F) Port X 1-11/16" OD (F-TA02)
Bulkhead Fitting, PTFE, 3/8" NPT(F) Port X 1-13/16" OD (F-TA03).
More Details
70618
results
<<<Previous
7
8
9
10
11
12
13
14
15
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Chemfluor Female Straight Couplers to Ptfe Bulkhead Unions
>
31125329-s-toc_2 31125329stoc2