Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 14
Nickel Plate Brass Manifolds
to
Luer Lock Fittings Chrome Plated Brass
Image
Product Detail
More Detail
Nickel Plate Brass Manifolds
Buy Nickel Plate Brass Manifolds Items Including:
Nickel Plated Brass Manifold, 3 male lock ports, 1 female luer port (YO-31507-91)
Nickel Plated Brass Manifold, 4 male lock ports, 1 female luer port (YO-31507-92)
Nickel Plated Brass Manifold, 5 male lock ports, 1 female luer port (YO-31507-93).
More Details
Manifold with Male Luer
Buy Manifold with Male Luer Items Including:
Nickel Plated Brass Manifold, 3 male lock ports, 1 female luer port (YO-31507-91)
Nickel Plated Brass Manifold, 4 male lock ports, 1 female luer port (YO-31507-92)
Nickel Plated Brass Manifold, 5 male lock ports, 1 female luer port (YO-31507-93).
More Details
Hose Manifold
Buy Hose Manifold Items Including:
Nickel Plated Brass Manifold, 3 male lock ports, 1 female luer port (YO-31507-91)
Nickel Plated Brass Manifold, 4 male lock ports, 1 female luer port (YO-31507-92)
Nickel Plated Brass Manifold, 5 male lock ports, 1 female luer port (YO-31507-93).
More Details
Unc Unf Npt Brass Fittings
Buy Unc Unf Npt Brass Fittings Items Including:
Luer fittings, Chrome-plated Luer adapters,female luer to 10-32 UNF th (YO-31507-71)
Luer fittings, Chrome-plated Luer adapters,female luer to 1/4-28 UNF t (YO-31507-72)
Compression pipe adapter, 1/8" NPT x 1/8", Brass, each (YO-31412-34).
More Details
1 Double Barbed Brass Fitting
Buy 1 Double Barbed Brass Fitting Items Including:
Nickel-plated brass fittings, male luer lock x 1/16" hose barb (YO-31507-62)
Nickel-plated brass fittings, female luer x 1/16" hose barb (YO-31507-64)
Coupling bodies; hose barb valved fittings; 1/4" ID; 2.00" length (LCD17004).
More Details
Brass Fitting for Natural Gas
Buy Brass Fitting for Natural Gas Items Including:
Nickel-plated brass fittings, female luer x male luer (YO-31507-50)
Nickel-plated brass fittings, male luer lock x male luer lock (YO-31507-55)
Pipe fittings, Nipple, Natural PP, 1/8" NPT, 1-3/32", 17/32", 1/4"; 10 (YO-30610-90).
More Details
Chrome Plated Steel Tubing
Buy Chrome Plated Steel Tubing Items Including:
Luer fittings, Chrome-plated Luer adapters,female luer to male luer lo (YO-31507-51)
Luer fittings, Chrome-plated Luer adapters,female luer plug (YO-31507-53)
Luer fittings, Chrome-plated Luer adapters,male luer lock to 1/8" to 3 (YO-31507-61).
More Details
Kar Products Brass Fittings 95378
Buy Kar Products Brass Fittings 95378 Items Including:
Luer fittings, Nickel-plated brass fitting, male slip x 1/4" barb (YO-31507-60)
Nickel-plated brass fittings, male luer lock x 1/16" hose barb (YO-31507-62)
Nickel-plated brass fittings, female luer x 1/16" hose barb (YO-31507-64).
More Details
Female Flare X Barb Brass Fitting
Buy Female Flare X Barb Brass Fitting Items Including:
Nickel-plated brass fittings, female luer x 1/16" hose barb (YO-31507-64)
Luer fittings, Nickel-plated brass fitting, male slip x 1/4" barb (YO-31507-60)
Rotating female luer to male luer lock adapter, Nickel-plated brass, e (YO-31507-80).
More Details
Brass Female Flare Fitting
Buy Brass Female Flare Fitting Items Including:
Nickel-plated brass fittings, female luer x 1/16" hose barb (YO-31507-64)
Rotating female luer to male luer lock adapter, Nickel-plated brass, e (YO-31507-80)
Luer fittings, nickel-plated brass three-way stopcocks, female luer x (YO-31507-11).
More Details
Galvanized Vs Nickel Plated Fittings 1
Buy Galvanized Vs Nickel Plated Fittings 1 Items Including:
Luer fittings, Nickel-plated brass fitting, male slip x 1/4" barb (YO-31507-60)
Nickel-plated brass fittings, male luer lock x 1/16" hose barb (YO-31507-62)
Nickel Plated Brass Manifold, 3 male lock ports, 1 female luer port (YO-31507-91).
More Details
High Pressure Hose Brass Fittings
Buy High Pressure Hose Brass Fittings Items Including:
Ultra-high pressure fittings; 10-32, STS male nut/ferrule; for 1/16" t (YO-01940-22)
Ultra-high pressure fittings; 10-32, STS male nut/ferrule; for 1/32" O (YO-01940-20)
Nickel-plated brass fittings, female luer x male luer (YO-31507-50).
More Details
Inch Brass Npt Fittings
Buy Inch Brass Npt Fittings Items Including:
Brass Sensor/Fittings; 350 psi max, sensor module brass 1/2"npt (423986F)
Brass Sensor/Fittings; 350 psi max, sensor module brass 3/4"npt (423 987 G)
Brass Sensor/Fittings; 350 psi max, sensor module brass 1.25"npt (423989J).
More Details
Brass Fitting Male Flange
Buy Brass Fitting Male Flange Items Including:
Luer fittings, Nickel-plated brass fitting, male slip x 1/4" barb (YO-31507-60)
Nickel-plated brass fittings, male luer lock x 10-32 UNF thread (YO-31507-70)
Rotating female luer to male luer lock adapter, Nickel-plated brass, e (YO-31507-80).
More Details
To 1 4 Reducer Brass Fitting
Buy To 1 4 Reducer Brass Fitting Items Including:
1/4" NPT brass female/male mini manual ball valve (MV608-4)
Compression To Thread Adapter, Stainless Steel, 6mm OD Tubing X 1/4" N (YO-31528-02)
Compression To Thread Adapter, Stainless Steel, 6mm OD Tubing X 1/4" N (YO-31528-24).
More Details
Brass Adapters Fittings
Buy Brass Adapters Fittings Items Including:
Threaded adapter, brass, 1/8" NPTM x 3/8" tubing ID, 5/pack (YO-30904-00)
Threaded adapter, brass, 1/8" NPTM x 1/4" tubing ID, 5/pack (YO-30904-01)
Metal quick-disconnect fittings, NPT (M) pipe adapters body,valved fit (MCD1004).
More Details
Nickel Plated Fittings E85
Buy Nickel Plated Fittings E85 Items Including:
Luer fittings, Nickel-plated brass fitting, male slip x 1/4" barb (YO-31507-60)
Nickel-plated brass fittings, male luer lock x 1/16" hose barb (YO-31507-62)
Nickel-plated brass fittings, female luer x 1/16" hose barb (YO-31507-64).
More Details
Brass Air Tube Fittings
Buy Brass Air Tube Fittings Items Including:
Rotating female luer to male luer lock adapter, Nickel-plated brass, e (YO-31507-80)
Luer fittings, Nickel-plated brass fitting, male slip x 1/4" barb (YO-31507-60)
Nickel-plated brass fittings, female luer x 1/16" hose barb (YO-31507-64).
More Details
Female Brass Fitting Vacuum
Buy Female Brass Fitting Vacuum Items Including:
Nickel-plated brass fittings, female luer x 1/16" hose barb (YO-31507-64)
Rotating female luer to male luer lock adapter, Nickel-plated brass, e (YO-31507-80)
Luer fittings, nickel-plated brass three-way stopcocks, female luer x (YO-31507-11).
More Details
Cole-Parmer Nickel-Plated Brass Fittings
Buy Cole-Parmer Nickel-Plated Brass Fittings Items Including:
Nickel Plated Brass Manifold, 3 male lock ports, 1 female luer port (YO-31507-91)
Nickel Plated Brass Manifold, 4 male lock ports, 1 female luer port (YO-31507-92)
Luer fittings, nickel-plated brass one-way stopcocks, female luer x m (YO-41507-00).
More Details
Nickel-Plated Brass Luer
Buy Nickel-Plated Brass Luer Items Including:
Nickel-plated brass fittings, male luer lock x male luer lock (YO-31507-55)
Rotating female luer to male luer lock adapter, Nickel-plated brass, e (YO-31507-80)
Luer fittings, nickel-plated brass one-way stopcocks, female luer x m (YO-31507-00).
More Details
Welding Hose Brass Fittings
Buy Welding Hose Brass Fittings Items Including:
Nickel-plated brass fittings, female luer x 1/16" hose barb (YO-31507-64)
Luer fittings, Nickel-plated brass fitting, male slip x 1/4" barb (YO-31507-60)
Rotating female luer to male luer lock adapter, Nickel-plated brass, e (YO-31507-80).
More Details
Male Brass Fittings
Buy Male Brass Fittings Items Including:
Luer fittings, Nickel-plated brass fitting, male slip x 1/4" barb (YO-31507-60)
Nickel-plated brass fittings, male luer lock x 10-32 UNF thread (YO-31507-70)
Luer fittings, Chrome-plated Luer adapters,female luer to male luer lo (YO-31507-51).
More Details
Alexandria Va Male Female Brass Fitting
Buy Alexandria Va Male Female Brass Fitting Items Including:
Rotating female luer to male luer lock adapter, Nickel-plated brass, e (YO-31507-80)
Nickel Plated Brass Manifold, 3 male lock ports, 1 female luer port (YO-31507-91)
Nickel Plated Brass Manifold, 4 male lock ports, 1 female luer port (YO-31507-92).
More Details
Brass Barb Adapter Fittings
Buy Brass Barb Adapter Fittings Items Including:
Threaded adapter, brass, 3/8" NPTM x 3/8" tubing ID, 50/pack (YO-30905-21)
Threaded adapter, brass, 3/8" NPTM x 1/2" tubing ID, 50/pack (YO-30905-22).
More Details
Female Brass Fitting 18
Buy Female Brass Fitting 18 Items Including:
Nickel-plated brass fittings, female luer x 1/16" hose barb (YO-31507-64)
Flowmeter, Panel-mount With Valve, With Brass Fittings; 18 SCFM O (YO-32605-54)
Rotating female luer to male luer lock adapter, Nickel-plated brass, e (YO-31507-80).
More Details
Nickel 60
Buy Nickel 60 Items Including:
Cole-Parmer Crucible Lid, Nickel; 15mL (1/pk)
Luer fittings, Nickel-plated brass fitting, male slip x 1/4" barb (YO-31507-60).
More Details
Brass Luer Fittings
Buy Brass Luer Fittings Items Including:
Luer fittings, Chrome-plated Luer adapters,female luer to male luer lo (YO-31507-51)
Luer fittings, Chrome-plated Luer adapters,female luer plug (YO-31507-53)
Luer fittings, Chrome-plated Luer adapters,male luer lock to 1/8" to 3 (YO-31507-61).
More Details
Inch Female Brass Fitting
Buy Inch Female Brass Fitting Items Including:
Nickel-plated brass fittings, female luer x 1/16" hose barb (YO-31507-64)
Rotating female luer to male luer lock adapter, Nickel-plated brass, e (YO-31507-80)
Luer fittings, nickel-plated brass three-way stopcocks, female luer x (YO-31507-11).
More Details
Luer Lock Fittings Chrome Plated Brass
Buy Luer Lock Fittings Chrome Plated Brass Items Including:
Rotating female luer to male luer lock adapter, Nickel-plated brass, e (YO-31507-80)
Luer fittings, Chrome-plated Luer adapters,male luer lock to 1/8" to 3 (YO-31507-61)
Luer fittings, Chrome-plated Luer adapters,female luer plug (YO-31507-53).
More Details
70618
results
<<<Previous
10
11
12
13
14
15
16
17
18
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Nickel Plate Brass Manifolds to Luer Lock Fittings Chrome Plated Brass
>
31125329-s-toc_2 31125329stoc2