Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 18
Ss Fitting Union 1
to
Metal Tube Fittings
Image
Product Detail
More Detail
Ss Fitting Union 1
Buy Ss Fitting Union 1 Items Including:
Parker 316 SS Compression union, 1/16" Tube OD (YO-30486-70)
316 SS fittings; female luer x 1/8" hose barb (YO-31507-28)
316 SS fittings; male luer lock x 1/4-28 UNF thread (YO-31507-32)
PTFE Fittings, Union connectors, elbow, 1 to 4 mm.
More Details
Male Luer Tubing Adapter
Buy Male Luer Tubing Adapter Items Including:
316 SS fittings; female luer x male luer lock (YO-31507-21)
316 SS fittings; male luer lock plug (YO-31507-23)
316 SS fittings; male luer lock x 1/4-28 UNF thread (YO-31507-32)
Nickel-plated brass fittings, female luer x male luer (YO-31507-50).
More Details
Luer Lock Fittings Mtll210 Jk1a
Buy Luer Lock Fittings Mtll210 Jk1a Items Including:
Snap luer lock ring, PC, 25/pk (YO-45504-74)
Snap luer lock ring, Nylon, 25/pk (YO-45505-74)
Nickel-plated brass fittings, male luer lock x male luer lock (YO-31507-55)
316 Stainless Steel fittings, male luer lock x male luer lock (YO-31507-90).
More Details
22 Degree Stainless Tubing
Buy 22 Degree Stainless Tubing Items Including:
Stainless Steel (SS) Hose Clamps, 3 1/2 x 4-3/8", box of 10
Winters Tube Stub Gauge, Stainless Steel, 2 1/2" Dial, Vacuum
THREADED SANITARY CLAMP ADAPTER, 316L SS, 1" CLAMP X 1" NPT( (YO-30560-22).
More Details
Tubing Fittings Male Thread to Hose
Buy Tubing Fittings Male Thread to Hose Items Including:
Barbed fitting kits, Threaded barb adapter kits, Natural PP, small fit (YO-31514-11)
Hose barb to male threaded adapter, Pharmafluorâ„¢ PTFE, 1/4" NPT(M) (YO-06362-50)
Nickel-plated brass fittings, male luer lock x 10-32 UNF thread (YO-31507-70).
More Details
Hose Barbs Male
Buy Hose Barbs Male Items Including:
Miniature Quick-Disconnect Hose Barb (Male), 1/16" ID (SMM0191)
Hose barb to male threaded adapter, Pharmafluorâ„¢ PTFE, 1/4" NPT(M) (YO-06362-50)
Luer fittings, 316 SS fittings, male luer lock x 1/8" hose barb (YO-31507-25).
More Details
Luer Tubing Barb
Buy Luer Tubing Barb Items Including:
Female luer to 1/16" low-profile semi-rigid tubing barb, PP, 25 per pa (YO-45500-12)
Female luer to 3/32" low-profile semi-rigid tubing barb, PP, 25 per pa (YO-45500-14).
More Details
Locking Luer Tubing
Buy Locking Luer Tubing Items Including:
316 SS fittings; female luer x male luer lock (YO-31507-21)
316 SS fittings; male luer lock x 1/4-28 UNF thread (YO-31507-32)
Male luer lock x 1/4" NPT, 316 SS, each (YO-31507-45)
Female luer lock x 1/8" NPT, 316 SS, each (YO-31507-46).
More Details
316 Ss Capillary Tubing
Buy 316 Ss Capillary Tubing Items Including:
316 SS Vacuum tubing, NW 16 flange, 10"
316 SS Vacuum tubing, NW 16 flange, 20"
316 SS fittings; female luer x male luer lock (YO-31507-21)
316 SS fittings; male slip plug (YO-31507-22)
316 SS fittings; male luer lock plug (YO-31507-23).
More Details
316 Coiled Ss Tubing
Buy 316 Coiled Ss Tubing Items Including:
316 SS tube mixer; 3/16" OD; 17 elements (YO-04669-50)
316 SS tube mixer; 3/16" OD; 21 elements (YO-04669-52)
316 SS tube mixer; 3/16" OD; 27 elements (YO-04669-54)
316 SS tube mixer; 1/4" OD; 21 elements (YO-04669-56).
More Details
Ss Tubing Tw Metals
Buy Ss Tubing Tw Metals Items Including:
316 SS Metering Valve with compression connections, 1/16" tube OD, 0.0 (1Z-NSL-V-SS-V)
316 SS Metering Valve with compression connections, 1/8" tube OD, 0.03 (2Z-NSL-V-SS-V)
Luer fittings, 316 SS fittings, female luer plug (YO-31507-24).
More Details
Steel Tubing 10 X 33
Buy Steel Tubing 10 X 33 Items Including:
Steel tubing clamp for up to 3/8"OD tubing (YO-06404-10)
Compression Straight Union, Stainless Steel, 6mm OD Tubing (YO-31528-33)
Poxygrid Wire Test Tube Half Rack 36 10-13mm (YO-06739-10)
Poxygrid Wire Test Tube Rack 72 10-13mm (F18750-0000).
More Details
22 Gage Stainless Tubing
Buy 22 Gage Stainless Tubing Items Including:
Stainless Steel (SS) Hose Clamps, 3 1/2 x 4-3/8", box of 10
Winters Tube Stub Gauge, Stainless Steel, 2 1/2" Dial, Vacuum
THREADED SANITARY CLAMP ADAPTER, 316L SS, 1" CLAMP X 1" NPT( (YO-30560-22).
More Details
Steel Luer Lock Fittings
Buy Steel Luer Lock Fittings Items Including:
316 SS fittings; female luer x male luer lock (YO-31507-21).
More Details
Luer Fittings Peek Tubing
Buy Luer Fittings Peek Tubing Items Including:
Plastic luer fittings, PVC tubing with luer ends, 48" L, M/F fitting e (YO-30600-61)
Luer fittings, Chrome-plated Luer adapters,female luer to male luer lo (YO-31507-51)
Luer fittings, 316 SS fittings, female luer x male slip (YO-31507-20).
More Details
Luer Lock Fittings in Georgia
Buy Luer Lock Fittings in Georgia Items Including:
Snap luer lock ring, PC, 25/pk (YO-45504-74)
Snap luer lock ring, Nylon, 25/pk (YO-45505-74)
Nickel-plated brass fittings, male luer lock x male luer lock (YO-31507-55)
Luer fittings, Chrome-plated Luer adapters,male luer lock plug (YO-31507-52).
More Details
Luer Lock Fittings Moldel
Buy Luer Lock Fittings Moldel Items Including:
Snap luer lock ring, PC, 25/pk (YO-45504-74)
Snap luer lock ring, Nylon, 25/pk (YO-45505-74)
Nickel-plated brass fittings, male luer lock x male luer lock (YO-31507-55)
Luer fittings, Chrome-plated Luer adapters,male luer lock plug (YO-31507-52).
More Details
Plastic Luer Lock Fittings
Buy Plastic Luer Lock Fittings Items Including:
Snap luer lock ring, PC, 25/pk (YO-45504-74)
Snap luer lock ring, Nylon, 25/pk (YO-45505-74)
Nickel-plated brass fittings, male luer lock x male luer lock (YO-31507-55)
Luer fittings, Chrome-plated Luer adapters,male luer lock plug (YO-31507-52).
More Details
Sealing Luer Lock Fittings Air
Buy Sealing Luer Lock Fittings Air Items Including:
Snap luer lock ring, PC, 25/pk (YO-45504-74)
Snap luer lock ring, Nylon, 25/pk (YO-45505-74)
Nickel-plated brass fittings, male luer lock x male luer lock (YO-31507-55)
316 Stainless Steel fittings, male luer lock x male luer lock (YO-31507-90).
More Details
Metal Luer Fittings
Buy Metal Luer Fittings Items Including:
Luer fittings, Chrome-plated Luer adapters,female luer to male luer lo (YO-31507-51)
Luer fittings, Chrome-plated Luer adapters,female luer plug (YO-31507-53)
Luer fittings, Chrome-plated Luer adapters,male luer lock to 1/8" to 3 (YO-31507-61).
More Details
Stainless Steel Tube Fitting Female
Buy Stainless Steel Tube Fitting Female Items Including:
316 SS fittings; female luer x male luer lock (YO-31507-21)
Luer fittings, 316 SS fittings, female luer plug (YO-31507-24)
316 SS fittings; female luer x 10-32 UNF thread (YO-31507-33)
316 SS fittings; female luer x 1/4-28 UNF thread (YO-31507-34).
More Details
Female Luer Lock Adapters
Buy Female Luer Lock Adapters Items Including:
Adapter, PP, 1/2" mini to female luer lock (YO-31200-50)
Adapter, Kynar, 1/2" mini to 1/2" to female luer lock
Female luer lock x 1/8" NPT, 316 SS, each (YO-31507-46)
Female luer lock x 1/4" NPT, 316 SS, each (YO-31507-48).
More Details
Male Luer Lock Tube Cap
Buy Male Luer Lock Tube Cap Items Including:
Nickel-plated brass fittings, male luer lock x male luer lock (YO-31507-55)
One-way stopcocks; male luer lock x male luer lock, nickel-plated bras (YO-31507-02)
Luer stopcock, 1-way, male luer lock x male luer lock, black PVDF, pac (YO-31200-90).
More Details
316 Ss Luer Fitting
Buy 316 Ss Luer Fitting Items Including:
Luer fittings, 316 SS fittings, female luer x male slip (YO-31507-20)
316 SS fittings; female luer x male luer lock (YO-31507-21).
More Details
Luer Lock Adapter 1 4 Npt
Buy Luer Lock Adapter 1 4 Npt Items Including:
Male luer lock x 1/4" NPT, 316 SS, each (YO-31507-45)
Female luer lock x 1/4" NPT, 316 SS, each (YO-31507-48)
Male luer lock x 1/4" NPT, Nickel-plated brass, each (YO-31507-85).
More Details
Cole-Parmer Metal Luer Fittings
Buy Cole-Parmer Metal Luer Fittings Items Including:
Luer fittings, Chrome-plated Luer adapters,female luer to male luer lo (YO-31507-51)
Luer fittings, Chrome-plated Luer adapters,female luer plug (YO-31507-53)
Luer fittings, Chrome-plated Luer adapters,male luer lock to 1/8" to 3 (YO-31507-61).
More Details
Ss Luer Fittings
Buy Ss Luer Fittings Items Including:
Luer fittings, 316 SS fittings, female luer x male slip (YO-31507-20).
More Details
Luer Fitting Tube Assemblies
Buy Luer Fitting Tube Assemblies Items Including:
Luer stopcock tubing assembly (YO-30526-04)
Plastic luer fittings, PVC tubing with luer ends, 48" L, M/F fitting e (YO-30600-61)
Plastic luer fittings, PVC tubing with luer ends, 24" L, M/M fitting e (YO-30600-62).
More Details
Tube Luer Fitting
Buy Tube Luer Fitting Items Including:
Plastic luer fittings, PVC tubing with luer ends, 48" L, M/F fitting e (YO-30600-61)
Luer fittings, Chrome-plated Luer adapters,female luer to male luer lo (YO-31507-51)
Luer fittings, Chrome-plated Luer adapters,female luer plug (YO-31507-53).
More Details
Metal Tube Fittings
Buy Metal Tube Fittings Items Including:
Metallic Tubing Cutter, 110-120/220-240 V (YO-95649-00)
Metal Quick-Disconnect NPT(F) Pipe Adapter Insert; Valved, 1/4" NPT(F) (MCD2604)
316 SS fittings; female luer x male luer lock (YO-31507-21)
316 SS fittings; male slip plug (YO-31507-22).
More Details
70618
results
<<<Previous
14
15
16
17
18
19
20
21
22
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Ss Fitting Union 1 to Metal Tube Fittings
>
31125329-s-toc_2 31125329stoc2