Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 19
Glass
to
Luer Lock Fittings
Image
Product Detail
More Detail
Glass to Luer Lock Fittings
Buy Glass to Luer Lock Fittings Items Including:
Snap luer lock ring, PC, 25/pk (YO-45504-74)
Snap luer lock ring, Nylon, 25/pk (YO-45505-74)
Nickel-plated brass fittings, male luer lock x male luer lock (YO-31507-55)
Luer fittings, Chrome-plated Luer adapters,male luer lock plug (YO-31507-52).
More Details
Luer Lock Fittings to Hypotube
Buy Luer Lock Fittings to Hypotube Items Including:
Snap luer lock ring, PC, 25/pk (YO-45504-74)
Snap luer lock ring, Nylon, 25/pk (YO-45505-74)
Nickel-plated brass fittings, male luer lock x male luer lock (YO-31507-55)
Luer fittings, Chrome-plated Luer adapters,male luer lock plug (YO-31507-52).
More Details
Luer Lock Fittings Charlotte Nc
Buy Luer Lock Fittings Charlotte Nc Items Including:
Snap luer lock ring, PC, 25/pk (YO-45504-74)
Snap luer lock ring, Nylon, 25/pk (YO-45505-74)
Nickel-plated brass fittings, male luer lock x male luer lock (YO-31507-55)
316 Stainless Steel fittings, male luer lock x male luer lock (YO-31507-90).
More Details
Luer Lock Fittings Grainger
Buy Luer Lock Fittings Grainger Items Including:
Snap luer lock ring, PC, 25/pk (YO-45504-74)
Snap luer lock ring, Nylon, 25/pk (YO-45505-74)
Nickel-plated brass fittings, male luer lock x male luer lock (YO-31507-55)
Luer fittings, Chrome-plated Luer adapters,male luer lock plug (YO-31507-52).
More Details
Luer Lock Fittings Retainer
Buy Luer Lock Fittings Retainer Items Including:
Snap luer lock ring, PC, 25/pk (YO-45504-74)
Snap luer lock ring, Nylon, 25/pk (YO-45505-74)
Nickel-plated brass fittings, male luer lock x male luer lock (YO-31507-55)
Luer fittings, Chrome-plated Luer adapters,male luer lock plug (YO-31507-52).
More Details
Luer Lock Fittings Tips
Buy Luer Lock Fittings Tips Items Including:
Snap luer lock ring, PC, 25/pk (YO-45504-74)
Snap luer lock ring, Nylon, 25/pk (YO-45505-74)
Nickel-plated brass fittings, male luer lock x male luer lock (YO-31507-55)
Luer fittings, Chrome-plated Luer adapters,male luer lock plug (YO-31507-52).
More Details
Medical Luer Lock Fittings
Buy Medical Luer Lock Fittings Items Including:
Snap luer lock ring, PC, 25/pk (YO-45504-74)
Snap luer lock ring, Nylon, 25/pk (YO-45505-74)
Nickel-plated brass fittings, male luer lock x male luer lock (YO-31507-55)
Luer fittings, Chrome-plated Luer adapters,male luer lock plug (YO-31507-52).
More Details
316 Steel Capillary Tubing
Buy 316 Steel Capillary Tubing Items Including:
316 Stainless Steel fittings, female luer x female luer (YO-31507-89)
Luer fittings, 316 SS fittings, female luer x male slip (YO-31507-20)
316 SS fittings; female luer x male luer lock (YO-31507-21)
316 SS fittings; male slip plug (YO-31507-22).
More Details
Luer Fittings Female Threads
Buy Luer Fittings Female Threads Items Including:
316 SS fittings; female luer x 10-32 UNF thread (YO-31507-33)
Nickel-plated brass fittings, female luer x female luer (YO-31507-56)
Female luer x female luer adapter, PC, 25/pk (YO-45501-22)
Female luer x female luer elbow, PC, 25/pk (YO-45501-26).
More Details
Female Metal Luer Lock Adapter
Buy Female Metal Luer Lock Adapter Items Including:
Rotating female luer to male luer lock adapter, Nickel-plated brass, e (YO-31507-80)
Luer fittings, 316 SS fittings, female luer lock x 1/16" hose barb (YO-31507-29)
3-way "Y" adapter, 1 female luer, 2 male luer lock (YO-31507-81).
More Details
Ss Tubing Fittings
Buy Ss Tubing Fittings Items Including:
Luer fittings, 316 SS fittings, female luer lock x 1/16" hose barb (YO-31507-29)
316 SS fittings; male slip plug (YO-31507-22)
316 SS fittings; male luer lock plug (YO-31507-23)
316 SS fittings; female luer x 1/8" hose barb (YO-31507-28).
More Details
Male Luer Lock Pressure Tubing
Buy Male Luer Lock Pressure Tubing Items Including:
Nickel-plated brass fittings, male luer lock x male luer lock (YO-31507-55)
One-way stopcocks; male luer lock x male luer lock, nickel-plated bras (YO-31507-02)
Luer stopcock, 1-way, male luer lock x male luer lock, black PVDF, pac (YO-31200-90).
More Details
Female Luer Lok Male Luer Slip
Buy Female Luer Lok Male Luer Slip Items Including:
Luer fittings, 316 SS fittings, female luer x male slip (YO-31507-20)
Male-female large-bore adapter, radiation-stable polycarbonate, pack o (YO-45509-91).
More Details
Metal Luer Adapters
Buy Metal Luer Adapters Items Including:
Rotating female luer to male luer lock adapter, Nickel-plated brass, e (YO-31507-80)
Luer fittings, Chrome-plated Luer adapters,female luer plug (YO-31507-53)
Luer fittings, Chrome-plated Luer adapters,male luer lock to 1/8" to 3 (YO-31507-61).
More Details
Metal Luer Lock 1 4 Fitting
Buy Metal Luer Lock 1 4 Fitting Items Including:
Luer fittings, Chrome-plated Luer adapters,male luer lock to 1/4" to 5 (YO-31507-66)
Luer fittings, Chrome-plated Luer adapters,male luer lock to 1/4-28 UN (YO-31507-73)
Male luer lock x 1/4" NPT, Nickel-plated brass, each (YO-31507-85).
More Details
Liquid Metal Tubing Fittings
Buy Liquid Metal Tubing Fittings Items Including:
Metallic Tubing Cutter, 110-120/220-240 V (YO-95649-00)
Metal-Detectable Gasket, Platinum-Cured Silicone, 2"
Luer fittings, 316 SS fittings, female luer x male slip (YO-31507-20)
Luer fittings, 316 SS fittings, female luer plug (YO-31507-24).
More Details
316 Ss Tubing
Buy 316 Ss Tubing Items Including:
316 SS tube mixer; 3/16" OD; 17 elements (YO-04669-50)
316 SS tube mixer; 3/16" OD; 21 elements (YO-04669-52)
316 SS tube mixer; 3/16" OD; 27 elements (YO-04669-54)
316 SS tube mixer; 1/4" OD; 21 elements (YO-04669-56).
More Details
Fitting 1 4 Hose to Threaded
Buy Fitting 1 4 Hose to Threaded Items Including:
316 SS fittings; male luer lock x 1/4-28 UNF thread (YO-31507-32)
Female luer bulkhead, 1/4-28 UNF thread, to 1/16" hose barb, PP, 25/pk (YO-45500-30)
Female luer bulkhead, 1/4-28 UNF thread, to 3/32" hose barb, PP, 25/pk (YO-45500-32).
More Details
Male Luer to Tube
Buy Male Luer to Tube Items Including:
316 SS fittings; female luer x male luer lock (YO-31507-21)
316 SS fittings; male luer lock plug (YO-31507-23)
316 SS fittings; male luer lock x 10-32 UNF thread (YO-31507-31)
316 SS fittings; male luer lock x 1/4-28 UNF thread (YO-31507-32).
More Details
Cole Luer Fittings and Adapters
Buy Cole Luer Fittings and Adapters Items Including:
Luer fittings, Polycarbonate fittings, female x male lock, rotating, (YO-06464-95)
Luer fittings, 316 SS fittings, male luer lock x 1/8" hose barb (YO-31507-25)
Luer fittings, 316 SS fittings, male luer lock x 1/4" hose barb (YO-31507-27).
More Details
Npt Luer Female
Buy Npt Luer Female Items Including:
Female luer lock x 1/8" NPT, 316 SS, each (YO-31507-46)
Female luer lock x 1/4" NPT, 316 SS, each (YO-31507-48)
Female luer x 1/8-27 NPT thread, Nylon, 25/pk (YO-45505-78)
Female luer x 1/8-27 NPT thread elbow, Nylon, 25/pk (YO-45505-80).
More Details
Cole 316 Ss Fittings
Buy Cole 316 Ss Fittings Items Including:
Luer fittings, 316 SS fittings, female luer x male slip (YO-31507-20)
Cole-Parmer Direct Reading Flowmeter with 316 SS Fittings and With Val (YO-32061-10)
Compression fitting; probe diameter, 3/16"; 316 SS; 1/8"NPT (M) (3-2 FH4BZ-SS-C).
More Details
Ga 316 Ss Tubing
Buy Ga 316 Ss Tubing Items Including:
316 SS tube mixer; 3/16" OD; 17 elements (YO-04669-50)
316 SS tube mixer; 3/16" OD; 21 elements (YO-04669-52)
316 SS tube mixer; 3/16" OD; 27 elements (YO-04669-54)
316 SS tube mixer; 1/4" OD; 21 elements (YO-04669-56).
More Details
4 Manderbent Ss Tubing
Buy 4 Manderbent Ss Tubing Items Including:
Barbed sanitary clamp adapter, 316L SS, 3/4" clamp x 1/4" tubing ID (YO-30560-09)
Barbed sanitary clamp adapter, 316L SS, 3/4" clamp x 3/4" tubing ID (YO-30560-12)
Stainless Steel (SS) Hose Clamps, 11/16 x 1-1/4", box of 10.
More Details
038 Wall 25 Ss Tubing
Buy 038 Wall 25 Ss Tubing Items Including:
High-head SS tube set for electric and air motors, 39"L
High-head SS tube set for electric and air motors, 47"L
High-head SS tube set for electric and air motors, 60"L
Stainless Steel (SS) Hose Clamps, 5 1/2 x 6-1/4", box of 10.
More Details
Luer Lock Female to Npt
Buy Luer Lock Female to Npt Items Including:
Female luer lock x 1/8" NPT, 316 SS, each (YO-31507-46)
Female luer lock x 1/4" NPT, 316 SS, each (YO-31507-48)
Female luer lock x 1/8" NPT, Nickel-plated brass, each (YO-31507-86).
More Details
Ss Tubing 25
Buy Ss Tubing 25 Items Including:
High-head SS tube set for electric and air motors, 39"L
High-head SS tube set for electric and air motors, 47"L
High-head SS tube set for electric and air motors, 60"L
Stainless Steel (SS) Hose Clamps, 5 1/2 x 6-1/4", box of 10.
More Details
Luer Lock Female for Tubings
Buy Luer Lock Female for Tubings Items Including:
Rotating female luer to male luer lock adapter, Nickel-plated brass, e (YO-31507-80)
Nickel Plated Brass Manifold, 3 male lock ports, 1 female luer port (YO-31507-91)
Nickel Plated Brass Manifold, 4 male lock ports, 1 female luer port (YO-31507-92).
More Details
4 Aluminized Steel Tubing
Buy 4 Aluminized Steel Tubing Items Including:
316 Stainless Steel Tubing; dimensions, 3/16" ID, 1/4" OD, 1/32" wall; (YO-03300-10)
304 Stainless Steel Tubing, 1/4" OD, 0.210" ID, 50ft roll (YO-34680-60)
Compression T-connector, 316 SS, 3/4" tubing OD (YO-31406-63).
More Details
27 Fitting
Buy 27 Fitting Items Including:
Luer fittings, 316 SS fittings, male luer lock x 1/4" hose barb (YO-31507-27)
Masterflex® straight barbed union, polypropylene, I/P 89, 10/pack (YO-30612-27)
Masterflex® barbed reducing union, polypropylene, I/P 89 x 1/2" tu (YO-30616-27).
More Details
70618
results
<<<Previous
15
16
17
18
19
20
21
22
23
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Glass to Luer Lock Fittings to 27 Fitting
>
31125329-s-toc_2 31125329stoc2