Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths & Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cleaning & Janitorial Supplies
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Lab Supplies & Consumables
Laboratory Equipment
Life Science
Masterflex® Peristaltic Pumps
Masterflex® Pump Tubing
Material Storage & Handling
Material Testing & Physical Testing
Microscopes & Magnification
Mixers and Blenders
pH, Conductivity, Dissolved Oxygen, and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity and Moisture Products
Tools
Tubing, Fittings, and Hose
Product Index Page 2
Ph Control Kit for Drinking Water
to
Gf Signet Ph
Image
Product Detail
More Detail
Ph Control Kit for Drinking Water
Buy Ph Control Kit for Drinking Water Items Including:
WTW ProfiLine pH 3110 drinking water set
WTW® ProfiLine pH 3210 drinking water set
WTW® ProfiLine pH 3310 drinking water set
Hach SensION™+ Drinking Water pH Meter Kit
YSI pH 100 kit with 3.3-ft (1-m) probe
More Details
Water Ph and Orp Meter
Buy Water Ph and Orp Meter Items Including:
Horiba D-55 pH/DO/ORP meter
(352735-1)
Eutech pH 6+ pH/ORP meter kit (YO-15940-80)
T-Fitting for In-Line Process pH and ORP Electrodes, Kynar (YO-27003-86)
Cole-parmer Preamplfied pH/orp Electrode Cable, No Atc Element, 25' (YO-27007-20).
More Details
Handheld Orp Meter
Buy Handheld Orp Meter Items Including:
Eutech pH 6+ pH/ORP meter kit (YO-15940-80).
More Details
Ph and Orp Meter for Aquariums
Buy Ph and Orp Meter for Aquariums Items Including:
Horiba D-55 pH/DO/ORP meter
(352735-1)
Eutech pH 6+ pH/ORP meter kit (YO-15940-80)
T-Fitting for In-Line Process pH and ORP Electrodes, Kynar (YO-27003-86)
Cole-parmer Preamplfied pH/orp Electrode Cable, No Atc Element, 25' (YO-27007-20).
More Details
Purified Water Ph Meter
Buy Purified Water Ph Meter Items Including:
Hach SensION™+ Drinking Water pH Meter Kit
Denver Instrument Portable Waterproof pH Meter
Pinnacle M530 Benchtop pH Meter, 115 VAC
Pinnacle M540 Benchtop pH Meter, 115 VAC
Pinnacle M545 Benchtop pH Meter, 115 VAC
More Details
Oakton Eutech Ph Meter
Buy Oakton Eutech Ph Meter Items Including:
Oakton® Waterproof pH 600 Meter with pH probe
Oakton® Waterproof pH 610 Meter with pH probe
Oakton® Acorn™ pH 5 meter kit (pH 5)
Oakton® Acorn™ pH 6 meter kit (pH 6)
Oakton® advanced pH 110 meter kit (pH 110)
More Details
Orp Meter for Drinking Water
Buy Orp Meter for Drinking Water Items Including:
WTW ProfiLine pH 3110 drinking water set
WTW® ProfiLine pH 3210 drinking water set
WTW® ProfiLine pH 3310 drinking water set
Horiba D-55 pH/DO/ORP meter
(352735-1)
More Details
6 in 1 Ph Meter
Buy 6 in 1 Ph Meter Items Including:
pH meter Russell with 3-in-1 probe
Eutech pH 5+ pH meter kit (YO-15940-79)
Oakton® Acorn™ pH 6 meter kit (pH 6)
Oakton® pH 6+ meter with probe
Oakton® pH 6+ meter only
More Details
Ph and Orp Meter
Buy Ph and Orp Meter Items Including:
Horiba D-55 pH/DO/ORP meter
(352735-1)
Eutech pH 6+ pH/ORP meter kit (YO-15940-80)
T-Fitting for In-Line Process pH and ORP Electrodes, Kynar (YO-27003-86)
Cole-parmer Preamplfied pH/orp Electrode Cable, No Atc Element, 25' (YO-27007-20).
More Details
Irrigation Water Ph Meter
Buy Irrigation Water Ph Meter Items Including:
Hach SensION™+ Drinking Water pH Meter Kit
Denver Instrument Portable Waterproof pH Meter
Pinnacle M530 Benchtop pH Meter, 115 VAC
Pinnacle M540 Benchtop pH Meter, 115 VAC
Pinnacle M545 Benchtop pH Meter, 115 VAC
More Details
Soil Water Ph Meter
Buy Soil Water Ph Meter Items Including:
Hach SensION™+ Drinking Water pH Meter Kit
Denver Instrument Portable Waterproof pH Meter
Pinnacle M530 Benchtop pH Meter, 115 VAC
Pinnacle M540 Benchtop pH Meter, 115 VAC
Pinnacle M545 Benchtop pH Meter, 115 VAC
More Details
Water Ph Meter Cheap
Buy Water Ph Meter Cheap Items Including:
Hach SensION™+ Drinking Water pH Meter Kit
Denver Instrument Portable Waterproof pH Meter
Pinnacle M530 Benchtop pH Meter, 115 VAC
Pinnacle M540 Benchtop pH Meter, 115 VAC
Pinnacle M545 Benchtop pH Meter, 115 VAC
More Details
Water Proof Ph Meter
Buy Water Proof Ph Meter Items Including:
Denver Instrument Portable Waterproof pH Meter
Hach SensION™+ Drinking Water pH Meter Kit
Pinnacle M530 Benchtop pH Meter, 115 VAC
Pinnacle M540 Benchtop pH Meter, 115 VAC
Pinnacle M545 Benchtop pH Meter, 115 VAC
More Details
The Ph Meter
Buy The Ph Meter Items Including:
Denver Instrument Portable Waterproof pH Meter
Pinnacle Scholar Benchtop pH Meter, 110 VAC
Pinnacle M530 Benchtop pH Meter, 115 VAC
Pinnacle M540 Benchtop pH Meter, 115 VAC
pH meter Russell with pH and ATC probes
More Details
Meters for Ph
Buy Meters for Ph Items Including:
Oakton® pH 700 Benchtop Meter, meter only (pH 700)
Oakton® advanced pH 110 meter only (pH 110)
Oakton® pH 300 meter kit (pH 300)
Oakton® pH 310 meter kit (pH 310)
Oakton® Waterproof pH 600 Meter with pH probe
More Details
Eutech Instruments Ph/Ion Handheld Meters
Buy Eutech Instruments Ph/Ion Handheld Meters Items Including:
Eutech pH 6+ pH/ORP meter kit (YO-15940-80).
More Details
Meter Instruments
Buy Meter Instruments Items Including:
Denver® benchtop pH/mV/temperature/ion/conductivity meter, 115 VAC
Denver® benchtop pH/mV/temperature/ion/conductivity meter, 230 VAC
Denver Instrument Portable Waterproof pH/mV/Ion Meter
Eutech Instruments Waterproof pH/DO Meter Kit (YO-99855-30).
More Details
Signet Ph Controller Mk710 3 Sensor
Buy Signet Ph Controller Mk710 3 Sensor Items Including:
+GF+® Signet pH/ORP Transmitter, field-mount with single input/dua
+GF+® Signet pH/ORP Transmitter, panel-mount with single input/dua
+GF+® Signet pH/ORP Transmitter, field-mount with relays
+GF+® Signet pH/ORP Transmitter, panel-mount with relays.
More Details
Industrial Junction Boxes
Buy Industrial Junction Boxes Items Including:
In-line sensor electronics w/junction box (3-2750-1)
In-line sensor electronics w/junction box and EasyCal (3-2750-2)
Junction Box Cable, 50 Feet.
More Details
Gf Signet Ph/Ion Electrodes
Buy Gf Signet Ph/Ion Electrodes Items Including:
+GF+ Signet DryLoc Connector with 3/4" NPT threads, 15-ft cable (3-2760-3)
GF Signet twist-Lock flat surface pH electrode - model 3-2714
Flat pH surface electrode, for use in trace HF (2% concentration)
Bulb ORP electrode with bulb protection.
More Details
Submersible Ph Sensor
Buy Submersible Ph Sensor Items Including:
Submersible Sensor electronics with 3/4" NPT threads, 15-ft cable (3-2750-3)
Submersible Sensor electronics with ISO threads, 15-ft cable (3-2750-4).
More Details
Electrode Assembly
Buy Electrode Assembly Items Including:
Replacement titration cell generator electrode assembly (300429.1)
Cole-Parmer® Removable Blue Glass Double-Junction pH Electrode, 10 (YO-27009-11)
Submersible pH Electrode, CPVC (YO-27102-00)
Submersible pH Electrode, CPVC, 100Ω RTD (YO-27102-10).
More Details
Ph Electronic
Buy Ph Electronic Items Including:
pH electronic, Digital
+GF+ Signet Conductivity Cell with Integral Electronics, K = 0.1, 4 to
Submersible Sensor electronics with 3/4" NPT threads, 15-ft cable (3-2750-3)
Submersible Sensor electronics with ISO threads, 15-ft cable (3-2750-4).
More Details
Ph and Orp Sensors
Buy Ph and Orp Sensors Items Including:
General purpose ORP sensor (YO-98765-12)
Submersible Sensor electronics with 3/4" NPT threads, 15-ft cable (3-2750-3)
Submersible Sensor electronics with ISO threads, 15-ft cable (3-2750-4)
In-line sensor electronics w/junction box (3-2750-1).
More Details
Ph Orp Signet
Buy Ph Orp Signet Items Including:
+GF+® Signet pH/ORP Transmitter, field-mount with single input/dua
+GF+® Signet pH/ORP Transmitter, panel-mount with single input/dua
+GF+® Signet pH/ORP Transmitter, field-mount with relays
+GF+® Signet pH/ORP Transmitter, panel-mount with relays.
More Details
Gf Signet Ph/Ion Industrial Electrodes
Buy Gf Signet Ph/Ion Industrial Electrodes Items Including:
pH Electrode, low conductivity, bulb, Pt-1000 ATC, ISO 7/1 R3/4 thread
pH Electrode, low conductivity, bulb, 3KOhm Balco thermistor, 3/4" NPT
ORP Electrode, flat surface, 3/4" NPT threads
ORP Electrode, flat surface, ISO 7/1 R3/4 threads.
More Details
Signet Ph Sensor
Buy Signet Ph Sensor Items Including:
Submersible Sensor electronics with 3/4" NPT threads, 15-ft cable (3-2750-3)
+GF+ Signet DryLoc Preamp with 3/4" NPT threads, 15-ft cable (3-2760-1)
+GF+ Signet DryLoc Connector with 3/4" NPT threads, 15-ft cable (3-2760-3).
More Details
Electronic Ph Sensor
Buy Electronic Ph Sensor Items Including:
Submersible Sensor electronics with 3/4" NPT threads, 15-ft cable (3-2750-3)
+GF+ Signet DryLoc Preamp with 3/4" NPT threads, 15-ft cable (3-2760-1)
+GF+ Signet DryLoc Connector with 3/4" NPT threads, 15-ft cable (3-2760-3).
More Details
Ph Sensor Connectors
Buy Ph Sensor Connectors Items Including:
+GF+ Signet DryLoc Connector with 3/4" NPT threads, 15-ft cable (3-2760-3).
More Details
Gf Signet Ph/Ion
Buy Gf Signet Ph/Ion Items Including:
+GF+® Signet pH/ORP Transmitter, field-mount with single input/dua
+GF+® Signet pH/ORP Transmitter, panel-mount with single input/dua
+GF+® Signet pH/ORP Transmitter, field-mount with relays
+GF+® Signet pH/ORP Transmitter, panel-mount with relays.
More Details
64361
results
<<<Previous
1
2
3
4
5
6
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,146
Ph Control Kit for Drinking Water to Gf Signet Ph
31125329-s-toc_2 31125329stoc2