Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 21
Cylindrical Plastic Tube Fittings Connectors
to
Reducing Plastic Fittings
Image
Product Detail
More Detail
Cylindrical Plastic Tube Fittings Connectors
Buy Cylindrical Plastic Tube Fittings Connectors Items Including:
Y Barbed Connector, 3/8, Polypropylene, Pack Of 100 (YO-31523-71)
Y Barbed Connector, 1", Polypropylene, Pack Of 10 (YO-31523-75)
T Barbed Connector, 1", Polypropylene, Pack Of 10 (YO-31523-80).
More Details
Dual Diameter Plastic Tube Fittings
Buy Dual Diameter Plastic Tube Fittings Items Including:
Plastic luer fittings, PVC tubing with luer ends, 48" L, M/F fitting e (YO-30600-61)
Plastic luer fittings, PVC tubing with luer ends, 24" L, M/M fitting e (YO-30600-62)
Straight Barbed Union, 3/8, Polypropylene, Pack Of 100 (YO-31523-60).
More Details
Lasco Plastic Hose Fittings
Buy Lasco Plastic Hose Fittings Items Including:
Plastic hose clamps, black, 0.569" x 0.650", 100/pack (YO-95613-08)
Plastic hose clamps, black, 0.859" x 0.989", 100/pack (YO-95613-13)
Plastic hose clamps, black, 1.123" x 1.296", 100/pack (YO-95613-27).
More Details
Mips to Barb Plastic Fitting
Buy Mips to Barb Plastic Fitting Items Including:
Straight Barbed Union With Lower-profile Barb, 3/8, Polycarbonate, Pac (YO-31523-95)
Reducing Barbed Union, 3/8x1/8, Polypropylene, Pack Of 100 (YO-31523-64)
Reducing Barbed Union, 3/8x1/4, Polypropylene, Pack Of 100 (YO-31523-65).
More Details
Value Plastics
Buy Value Plastics Items Including:
Barbed to sanitary adapter, 1/4" to mini sanitary, animal-free polypro (YO-31513-51)
Barbed to sanitary adapter, 3/4" to mini sanitary, animal-free polypro (YO-31513-54)
Straight Barbed Union, 3/8, Polypropylene, Pack Of 100 (YO-31523-60).
More Details
Value Plastics Barbed to Barbed Connectors
Buy Value Plastics Barbed to Barbed Connectors Items Including:
Straight Barbed Union, 3/8, Polypropylene, Pack Of 100 (YO-31523-60)
Straight Barbed Union, 1/2, Polypropylene, Pack Of 100 (YO-31523-61)
Reducing Barbed Union, 3/8x1/4, Animal-derived Ingredient-free Polypro (YO-31526-41).
More Details
Plastic Tube Fittings Instruction
Buy Plastic Tube Fittings Instruction Items Including:
Plastic luer fittings, PVC tubing with luer ends, 48" L, M/F fitting e (YO-30600-61)
Plastic luer fittings, PVC tubing with luer ends, 24" L, M/M fitting e (YO-30600-62)
Straight Barbed Union, 3/8, Polypropylene, Pack Of 100 (YO-31523-60).
More Details
Plastic Tube Fittings Semiconductor
Buy Plastic Tube Fittings Semiconductor Items Including:
Plastic luer fittings, PVC tubing with luer ends, 48" L, M/F fitting e (YO-30600-61)
Plastic luer fittings, PVC tubing with luer ends, 24" L, M/M fitting e (YO-30600-62)
Straight Barbed Union, 3/8, Polypropylene, Pack Of 100 (YO-31523-60).
More Details
Plastic Tube Fittings Water
Buy Plastic Tube Fittings Water Items Including:
Plastic luer fittings, PVC tubing with luer ends, 48" L, M/F fitting e (YO-30600-61)
Plastic luer fittings, PVC tubing with luer ends, 24" L, M/M fitting e (YO-30600-62)
Straight Barbed Union, 3/8, Polypropylene, Pack Of 100 (YO-31523-60).
More Details
Serrated Plastic Tube Fittings
Buy Serrated Plastic Tube Fittings Items Including:
Plastic luer fittings, PVC tubing with luer ends, 48" L, M/F fitting e (YO-30600-61)
Plastic luer fittings, PVC tubing with luer ends, 24" L, M/M fitting e (YO-30600-62)
Straight Barbed Union, 3/8, Polypropylene, Pack Of 100 (YO-31523-60).
More Details
Variuos Plastic Tube Fittings
Buy Variuos Plastic Tube Fittings Items Including:
Plastic luer fittings, PVC tubing with luer ends, 48" L, M/F fitting e (YO-30600-61)
Plastic luer fittings, PVC tubing with luer ends, 24" L, M/M fitting e (YO-30600-62)
Straight Barbed Union, 3/8, Polypropylene, Pack Of 100 (YO-31523-60).
More Details
Value Plastics Large-Bore Barbed Connectors
Buy Value Plastics Large-Bore Barbed Connectors Items Including:
Reducing Barbed Union, 3/8x1/8, Polypropylene, Pack Of 100 (YO-31523-64)
Reducing Barbed Union, 3/8x1/4, Polypropylene, Pack Of 100 (YO-31523-65)
Reducing Barbed Union, 1/2x1/4, Polypropylene, Pack Of 100 (YO-31523-66).
More Details
Small Plastic Hose Fittings
Buy Small Plastic Hose Fittings Items Including:
Plastic hose clamps, black, 0.569" x 0.650", 100/pack (YO-95613-08)
Barbed fitting kits, Barbed connector/reducer kits, PP, small fittings (YO-31514-03)
Barbed fitting kits, Threaded barb adapter kits, Natural PP, small fit (YO-31514-11).
More Details
Value Plastics Barbed Fittings
Buy Value Plastics Barbed Fittings Items Including:
Barbed to sanitary adapter, 1/4" to mini sanitary, animal-free polypro (YO-31513-51)
Barbed to sanitary adapter, 3/4" to mini sanitary, animal-free polypro (YO-31513-54)
Straight Barbed Union, 3/8, Polypropylene, Pack Of 100 (YO-31523-60).
More Details
Plastic Tubing Barb Connector
Buy Plastic Tubing Barb Connector Items Including:
Y Barbed Connector, 3/8, Polypropylene, Pack Of 100 (YO-31523-71)
Y Barbed Connector, 1", Polypropylene, Pack Of 10 (YO-31523-75)
T Barbed Connector, 1", Polypropylene, Pack Of 10 (YO-31523-80).
More Details
Value Plastics Tubing, Fittings, and Hose
Buy Value Plastics Tubing, Fittings, and Hose Items Including:
Barbed to sanitary adapter, 1/4" to mini sanitary, animal-free polypro (YO-31513-70)
Reducing Barbed Union, 3/8x1/8, Polycarbonate, Pack Of 100 (YO-31523-84)
Reducing Barbed Union, 3/8x1/4, Polycarbonate, Pack Of 100 (YO-31523-85).
More Details
Mcmaster Barbed Plastic Fitting
Buy Mcmaster Barbed Plastic Fitting Items Including:
Straight Barbed Union With Lower-profile Barb, 3/8, Polycarbonate, Pac (YO-31523-95)
Reducing Barbed Union, 3/8x1/8, Polypropylene, Pack Of 100 (YO-31523-64)
Reducing Barbed Union, 3/8x1/4, Polypropylene, Pack Of 100 (YO-31523-65).
More Details
Plastic Tubing Connectors Fitting
Buy Plastic Tubing Connectors Fitting Items Including:
Y Barbed Connector, 3/8, Polypropylene, Pack Of 100 (YO-31523-71)
Y Barbed Connector, 1/2, Polypropylene, Pack Of 100 (YO-31523-72)
Straight Barbed Union, 3/8, Animal-derived Ingredient-free Polypropyle (YO-31526-40).
More Details
Plastic Hose Fittings Connectors
Buy Plastic Hose Fittings Connectors Items Including:
Plastic hose clamps, black, 0.569" x 0.650", 100/pack (YO-95613-08)
Plastic hose clamps, black, 0.859" x 0.989", 100/pack (YO-95613-13)
Plastic hose clamps, black, 1.123" x 1.296", 100/pack (YO-95613-27).
More Details
Clear Plastic Tube Fittings
Buy Clear Plastic Tube Fittings Items Including:
Plastic luer fittings, PVC tubing with luer ends, 48" L, M/F fitting e (YO-30600-61)
Plastic hose clamps, black, 0.569" x 0.650", 100/pack (YO-95613-08)
Plastic hose clamps, black, 0.859" x 0.989", 100/pack (YO-95613-13).
More Details
Imperial Plastic Tube Fittings
Buy Imperial Plastic Tube Fittings Items Including:
Plastic luer fittings, PVC tubing with luer ends, 48" L, M/F fitting e (YO-30600-61)
Plastic luer fittings, PVC tubing with luer ends, 24" L, M/M fitting e (YO-30600-62)
Straight Barbed Union, 3/8, Polypropylene, Pack Of 100 (YO-31523-60).
More Details
Long Plastic Tube Fittings
Buy Long Plastic Tube Fittings Items Including:
Plastic luer fittings, PVC tubing with luer ends, 48" L, M/F fitting e (YO-30600-61)
Plastic luer fittings, PVC tubing with luer ends, 24" L, M/M fitting e (YO-30600-62)
Straight Barbed Union, 3/8, Polypropylene, Pack Of 100 (YO-31523-60).
More Details
Mtric Plastic Tube Fittings
Buy Mtric Plastic Tube Fittings Items Including:
Plastic luer fittings, PVC tubing with luer ends, 48" L, M/F fitting e (YO-30600-61)
Plastic luer fittings, PVC tubing with luer ends, 24" L, M/M fitting e (YO-30600-62)
Straight Barbed Union, 3/8, Polypropylene, Pack Of 100 (YO-31523-60).
More Details
Connectors for Plastic Tubings
Buy Connectors for Plastic Tubings Items Including:
Y Barbed Connector, 3/8, Polypropylene, Pack Of 100 (YO-31523-71)
Y Barbed Connector, 1", Polypropylene, Pack Of 10 (YO-31523-75)
T Barbed Connector, 1", Polypropylene, Pack Of 10 (YO-31523-80).
More Details
Value Barbed to Barbed Connectors
Buy Value Barbed to Barbed Connectors Items Including:
Reducing Barbed Union, 3/8x1/8, Polycarbonate, Pack Of 100 (YO-31523-84)
Reducing Barbed Union, 3/8x1/4, Polycarbonate, Pack Of 100 (YO-31523-85)
Reducing Barbed Union, 1/2x1/4, Polycarbonate, Pack Of 100 (YO-31523-86).
More Details
Hose Fittings Plastic Tubing
Buy Hose Fittings Plastic Tubing Items Including:
Barbed to sanitary adapter, 1/4" to mini sanitary, animal-free polypro (YO-31513-70)
Reducing Barbed Union, 3/8x1/8, Polypropylene, Pack Of 100 (YO-31523-64)
Reducing Barbed Union, 3/8x1/4, Polypropylene, Pack Of 100 (YO-31523-65).
More Details
Barb Plastic
Buy Barb Plastic Items Including:
Barbed to sanitary adapter, 1/4" to mini sanitary, animal-free polypro (YO-31513-51)
Barbed to sanitary adapter, 1/4" to mini sanitary, PVDF (YO-31513-61)
Barbed to sanitary adapter, 1/4" to 1-1/2" sanitary, PVDF (YO-31513-65).
More Details
Plastic Hose Connectors
Buy Plastic Hose Connectors Items Including:
Reducing Barbed Union, 3/8x1/8, Polypropylene, Pack Of 100 (YO-31523-64)
Reducing Barbed Union, 3/8x1/4, Polypropylene, Pack Of 100 (YO-31523-65)
Reducing Barbed Union, 1/2x1/4, Polypropylene, Pack Of 100 (YO-31523-66).
More Details
Union Plastic Fittings
Buy Union Plastic Fittings Items Including:
Reducing Barbed Union, 1x3/4, Polycarbonate, Pack Of 10 (YO-31523-90)
Straight Barbed Union With Lower-profile Barb, 3/8, Polycarbonate, Pac (YO-31523-95)
Straight Barbed Union, 3/8, Animal-derived Ingredient-free Polypropyle (YO-31526-40).
More Details
Reducing Plastic Fittings
Buy Reducing Plastic Fittings Items Including:
Reducing Barbed Union, 3/8x1/8, Polypropylene, Pack Of 100 (YO-31523-64)
Reducing Barbed Union, 3/8x1/4, Polypropylene, Pack Of 100 (YO-31523-65)
Reducing Barbed Union, 1/2x1/4, Polypropylene, Pack Of 100 (YO-31523-66).
More Details
70618
results
<<<Previous
17
18
19
20
21
22
23
24
25
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
40
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Cylindrical Plastic Tube Fittings Connectors to Reducing Plastic Fittings
>
31125329-s-toc_2 31125329stoc2