Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 23
Tygon Tubing Material
to
Sanitary Clamps
Image
Product Detail
More Detail
Tygon Tubing Material
Buy Tygon Tubing Material Items Including:
Tygon® high-purity tubing, 1/16"ID x 3/16"OD, 50 ft/pack (YO-95635-00)
Tygon® high-purity tubing, 1/8"ID x 1/4"OD, 50 ft/pack (YO-95635-01).
More Details
Tygon Tubing for Bioprocessing
Buy Tygon Tubing for Bioprocessing Items Including:
Tygon® high-purity tubing, 1/16"ID x 3/16"OD, 50 ft/pack (YO-95635-00)
Tygon® high-purity tubing, 1/8"ID x 1/4"OD, 50 ft/pack (YO-95635-01)
Tygon® high-purity tubing, 5/16"ID x 7/16"OD, 50 ft/pack (YO-95635-04).
More Details
Flexible Tubing Material
Buy Flexible Tubing Material Items Including:
Tygon® high-purity tubing, 1/16"ID x 3/16"OD, 50 ft/pack (YO-95635-00)
Tygon® high-purity tubing, 1/8"ID x 1/4"OD, 50 ft/pack (YO-95635-01).
More Details
High Strength Nylon Tubing Mm
Buy High Strength Nylon Tubing Mm Items Including:
Barbed Fittings; Cross Connector, Nylon, 5 mm Tubing ID, 50/pk (YO-31418-11)
Tygon® high-purity tubing, 1/16"ID x 3/16"OD, 50 ft/pack (YO-95635-00)
Tygon® high-purity tubing, 1/8"ID x 1/4"OD, 50 ft/pack (YO-95635-01).
More Details
Tygon High-Purity Tubing, Biopharma
Buy Tygon High-Purity Tubing, Biopharma Items Including:
Tygon® high-purity tubing, 1/16"ID x 3/16"OD, 50 ft/pack (YO-95635-00)
Tygon® high-purity tubing, 1/8"ID x 1/4"OD, 50 ft/pack (YO-95635-01)
Tygon® high-purity tubing, 5/16"ID x 7/16"OD, 50 ft/pack (YO-95635-04).
More Details
Tygon Single-Use Technology
Buy Tygon Single-Use Technology Items Including:
Tygon® high-purity tubing, 1/16"ID x 3/16"OD, 50 ft/pack (YO-95635-00)
Tygon® high-purity tubing, 1/8"ID x 1/4"OD, 50 ft/pack (YO-95635-01)
Tygon® high-purity tubing, 5/16"ID x 7/16"OD, 50 ft/pack (YO-95635-04).
More Details
Bioprocess Technology
Buy Bioprocess Technology Items Including:
Tygon® high-purity tubing, 1/16"ID x 3/16"OD, 50 ft/pack (YO-95635-00)
Tygon® high-purity tubing, 1/8"ID x 1/4"OD, 50 ft/pack (YO-95635-01)
Tygon® high-purity tubing, 5/16"ID x 7/16"OD, 50 ft/pack (YO-95635-04).
More Details
High Purity Plastic Fittings
Buy High Purity Plastic Fittings Items Including:
Tygon® high-purity tubing, 1/16"ID x 3/16"OD, 50 ft/pack (YO-95635-00)
Tygon® high-purity tubing, 1/8"ID x 1/4"OD, 50 ft/pack (YO-95635-01)
Tygon® high-purity tubing, 5/16"ID x 7/16"OD, 50 ft/pack (YO-95635-04).
More Details
Single-Use Technologies
Buy Single-Use Technologies Items Including:
Tygon® high-purity tubing, 1/16"ID x 3/16"OD, 50 ft/pack (YO-95635-00)
Tygon® high-purity tubing, 1/8"ID x 1/4"OD, 50 ft/pack (YO-95635-01)
Tygon® high-purity tubing, 5/16"ID x 7/16"OD, 50 ft/pack (YO-95635-04).
More Details
Biopharma Fittings
Buy Biopharma Fittings Items Including:
PendoTECH PressureMAT Single-Use Sensor, Luer Fitting, Polycarbonate (PRESS-S-000)
Tygon® high-purity tubing, 1/16"ID x 3/16"OD, 50 ft/pack (YO-95635-00)
Tygon® high-purity tubing, 5/16"ID x 7/16"OD, 50 ft/pack (YO-95635-04).
More Details
L/S Biopharma
Buy L/S Biopharma Items Including:
Tygon® high-purity tubing, 1/8"ID x 1/4"OD, 50 ft/pack (YO-95635-01)
PendoTECH PressureMAT Single-Use Sensor, Polysulfone Luer Fitting (PREPS-N-000)
PendoTECH PressureMAT Single-Use Sensor, 1/2" Polysulphone Hose Barb F (PREPS-N-050).
More Details
Gaskets for Ansi 150# Flanges
Buy Gaskets for Ansi 150# Flanges Items Including:
ANSI 150# Ff Gasket,epdm,1/2" (YO-30549-01).
More Details
Ansi Gasket
Buy Ansi Gasket Items Including:
ANSI 150# Ff Gasket,epdm,1/2" (YO-30549-01).
More Details
Ansi Flange Gasket
Buy Ansi Flange Gasket Items Including:
ANSI 150# Ff Gasket,epdm,1/2" (YO-30549-01).
More Details
304l Stainless Steel Tubing
Buy 304l Stainless Steel Tubing Items Including:
SANITARY CLAMP, 304L SS, 1/2 - 3/4" (YO-30562-01)
SANITARY CLAMP, 304L SS, 2" (YO-30562-03)
Sanitary Clamp, 304L SS, 2-1/2" (YO-30562-04)
Sanitary Clamp, 304L SS, 3" (YO-30562-05)
Sanitary Clamp, 304L SS, 4" (YO-30562-06).
More Details
Stainless Steel Tubing with Sanitary Fittings
Buy Stainless Steel Tubing with Sanitary Fittings Items Including:
SANITARY CLAMP, 304L SS, 1/2 - 3/4" (YO-30562-01)
SANITARY CLAMP, 304L SS, 2" (YO-30562-03)
Sanitary Clamp, 304L SS, 2-1/2" (YO-30562-04)
Sanitary Clamp, 304L SS, 3" (YO-30562-05)
Sanitary Clamp, 304L SS, 4" (YO-30562-06).
More Details
Steel 01
Buy Steel 01 Items Including:
Paddle Wheel Flow Meter, Stainless Steel, 0.1 to 10 GPM (SES-050)
Concentric Sanitary Clamp Reducer, 316L SS, 1-1/2" X 1" (YO-30560-01)
Stainless steel cylindrical utility tank, 12 quart
Stainless steel cylindrical utility tank, 20 quart.
More Details
Stainless Tubing Clamp Fittings
Buy Stainless Tubing Clamp Fittings Items Including:
Tee Fittings, 63 ISO-K, 316 SS (C100-07-207)
Cross Fittings, 63 ISO-K, 316 SS (C100-07-211)
Cross Fitting, 100 ISO-K, 316 SS (C100-09-211)
SANITARY CLAMP, 304L SS, 1" - 1-1/2" (YO-30562-02)
SANITARY CLAMP, 304L SS, 2" (YO-30562-03).
More Details
304l Stainless
Buy 304l Stainless Items Including:
SANITARY CLAMP, 304L SS, 1/2 - 3/4" (YO-30562-01)
SANITARY CLAMP, 304L SS, 2" (YO-30562-03)
Sanitary Clamp, 304L SS, 2-1/2" (YO-30562-04)
Sanitary Clamp, 304L SS, 3" (YO-30562-05)
Sanitary Clamp, 304L SS, 4" (YO-30562-06).
More Details
304l Ss
Buy 304l Ss Items Including:
SANITARY CLAMP, 304L SS, 1/2 - 3/4" (YO-30562-01)
SANITARY CLAMP, 304L SS, 2" (YO-30562-03)
Sanitary Clamp, 304L SS, 2-1/2" (YO-30562-04)
Sanitary Clamp, 304L SS, 3" (YO-30562-05)
Sanitary Clamp, 304L SS, 4" (YO-30562-06).
More Details
2 Threaded Coupling and Rmc
Buy 2 Threaded Coupling and Rmc Items Including:
Straight threaded coupling, nylon, 1/2" NPT(F), pack of 6 (YO-06349-03)
Straight Union Coupling, PTFE, 1/2" NPT(F) (F-SC04)
316 SS Straight Coupling, 1/2" NPT(F) (YO-31410-03)
Reducing Coupling, PTFE, 3/4x1/2" NPT(F) (F-RC0604).
More Details
Threaded Sanitary Fittings
Buy Threaded Sanitary Fittings Items Including:
Cole-Parmer, Threaded Sanitary Clamp Adapter, 304L SS, 1" Clamp X 3/4"
THREADED SANITARY CLAMP ADAPTER, 316L SS, 3/4" CLAMP X 1/8" (YO-30560-16)
THREADED SANITARY CLAMP ADAPTER, 316L SS, 3/4" CLAMP X 1/4" (YO-30560-17).
More Details
Threaded Hose Clamp
Buy Threaded Hose Clamp Items Including:
Cole-Parmer, Threaded Sanitary Clamp Adapter, 316L SS, 1.5" Clamp X 3/
THREADED SANITARY CLAMP ADAPTER, 316L SS, 1" CLAMP X 3/4" NP (YO-30560-21)
THREADED SANITARY CLAMP ADAPTER, 316L SS, 1-1/2" CLAMP X 3/4" (YO-30560-25).
More Details
Threaded by Hose Adapter Fittings
Buy Threaded by Hose Adapter Fittings Items Including:
Compression To Thread Adapter, Stainless Steel, 10mm OD Tubing X 1/2" (YO-31528-30)
THREADED SANITARY CLAMP ADAPTER, 316L SS, 1-1/2" CLAMP X 3/4" (YO-30560-25).
More Details
Sanitary Fittings for Hose
Buy Sanitary Fittings for Hose Items Including:
Sanitary fittings, Plastic sanitary couplings, PVDF,1/4" ID, 1-9/16", (YO-31512-03)
ECCENTRIC SANITARY CLAMP REDUCER, 316L SS, 1-1/2" X 1" (YO-30560-05)
ECCENTRIC SANITARY CLAMP REDUCER, 316L SS, 2" X 1" (YO-30560-06).
More Details
316l Stainless Steel Threaded Adapters
Buy 316l Stainless Steel Threaded Adapters Items Including:
THREADED SANITARY CLAMP ADAPTER, 316L SS, 1-1/2" CLAMP X 3/4" (YO-30560-25)
THREADED SANITARY CLAMP ADAPTER, 316L SS, 1-1/2" CLAMP X 1-1/4&qu (YO-30560-27).
More Details
Sanitary Stainless Fittings Tube
Buy Sanitary Stainless Fittings Tube Items Including:
ECCENTRIC SANITARY CLAMP REDUCER, 316L SS, 1" X 3/4" (YO-30560-04)
ECCENTRIC SANITARY CLAMP REDUCER, 316L SS, 1-1/2" X 1" (YO-30560-05)
Barbed sanitary clamp adapter, 316L SS, 3/4" clamp x 1/8" tubing ID (YO-30560-08).
More Details
4 Threaded Steel Couplings
Buy 4 Threaded Steel Couplings Items Including:
Adapter,1/4"VCR(M) x NW16, 316LSS (C105-01-108)
316 SS Straight Coupling, 1/4" NPT(F) (YO-31410-01)
316 SS Straight Coupling, 3/4" NPT(F) (YO-31410-04)
Cole-Parmer, Threaded Sanitary Clamp Adapter, 304L SS, 1" Clamp X 3/4".
More Details
Sanitary Steel Tubing
Buy Sanitary Steel Tubing Items Including:
ECCENTRIC SANITARY CLAMP REDUCER, 316L SS, 1" X 3/4" (YO-30560-04)
ECCENTRIC SANITARY CLAMP REDUCER, 316L SS, 1-1/2" X 1" (YO-30560-05)
Barbed sanitary clamp adapter, 316L SS, 3/4" clamp x 1/8" tubing ID (YO-30560-08).
More Details
Sanitary Clamps
Buy Sanitary Clamps Items Including:
Concentric Sanitary Clamp Reducer, 316L SS, 1-1/2" X 1" (YO-30560-01)
ECCENTRIC SANITARY CLAMP REDUCER, 316L SS, 1" X 3/4" (YO-30560-04)
Barbed sanitary clamp adapter, 316L SS, 3/4" clamp x 1/8" tubing ID (YO-30560-08).
More Details
70618
results
<<<Previous
19
20
21
22
23
24
25
26
27
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
36
37
40
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Tygon Tubing Material to Sanitary Clamps
>
31125329-s-toc_2 31125329stoc2