Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 24
Steel Tube Couplings
to
Carbon Steel Tubing Elbows
Image
Product Detail
More Detail
Steel Tube Couplings
Buy Steel Tube Couplings Items Including:
Barbed sanitary clamp adapter, 316L SS, 3/4" clamp x 1/8" tubing ID (YO-30560-08)
Barbed sanitary clamp adapter, 316L SS, 1-1/2" clamp x 1-1/2" tubing I (YO-30560-14).
More Details
Sanitary Tube Clamps
Buy Sanitary Tube Clamps Items Including:
Barbed to sanitary adapter, 1/4" to mini sanitary, animal-free polypro (YO-31513-70)
ECCENTRIC SANITARY CLAMP REDUCER, 316L SS, 1-1/2" X 1" (YO-30560-05)
ECCENTRIC SANITARY CLAMP REDUCER, 316L SS, 2" X 1-1/2" (YO-30560-07).
More Details
Hose Threaded Couplings
Buy Hose Threaded Couplings Items Including:
Pipe fittings, Straight coupling, Chemfluor® PTFE, 1/8" NPT(F), 15 (YO-06469-16)
Pipe fittings, Straight coupling, Chemfluor® PTFE, 1/2" NPT(F), 1- (YO-06469-22).
More Details
Sanitary Hoses
Buy Sanitary Hoses Items Including:
ECCENTRIC SANITARY CLAMP REDUCER, 316L SS, 1-1/2" X 1" (YO-30560-05)
ECCENTRIC SANITARY CLAMP REDUCER, 316L SS, 2" X 1" (YO-30560-06)
Barbed sanitary clamp adapter, 316L SS, 1-1/2" clamp x 1-1/2" tubing I (YO-30560-14).
More Details
Tubing Coupling Adapter
Buy Tubing Coupling Adapter Items Including:
Keck Tubing Coupling Adapter, 10/pk
Barbed sanitary clamp adapter, 316L SS, 3/4" clamp x 1/8" tubing ID (YO-30560-08)
Barbed sanitary clamp adapter, 316L SS, 1-1/2" clamp x 1-1/2" tubing I (YO-30560-14).
More Details
Stainless Steel Sanitary Hose Fitting
Buy Stainless Steel Sanitary Hose Fitting Items Including:
Cole-Parmer, Threaded Sanitary Clamp Adapter, 316L SS, 1.5" Clamp X 3/
THREADED SANITARY CLAMP ADAPTER, 316L SS, 1" CLAMP X 1-1/2" (YO-30560-23)
THREADED SANITARY CLAMP ADAPTER, 316L SS, 1-1/2" CLAMP X 1-1/4&qu (YO-30560-27).
More Details
Stainless Sanitary 4 Tubing
Buy Stainless Sanitary 4 Tubing Items Including:
ECCENTRIC SANITARY CLAMP REDUCER, 316L SS, 1" X 3/4" (YO-30560-04)
THREADED SANITARY CLAMP ADAPTER, 316L SS, 3/4" CLAMP X 1/8" (YO-30560-16)
THREADED SANITARY CLAMP ADAPTER, 316L SS, 3/4" CLAMP X 1/4" (YO-30560-17).
More Details
Sanitary Tube Sanitary Tube Stainless
Buy Sanitary Tube Sanitary Tube Stainless Items Including:
ECCENTRIC SANITARY CLAMP REDUCER, 316L SS, 1" X 3/4" (YO-30560-04)
THREADED SANITARY CLAMP ADAPTER, 316L SS, 1" CLAMP X 2" NPT( (YO-30560-24)
THREADED SANITARY CLAMP ADAPTER, 316L SS, 1-1/2" CLAMP X 3/4" (YO-30560-25).
More Details
Steel Tee
Buy Steel Tee Items Including:
Cr4T010 Stainless Steel Tee 1.0 (CR4T010)
Cr4T015 Stainless Steel Tee 1.5 (CR4T015)
SANITARY CLAMP TEE, 316L SS, 3/4" (YO-30560-38)
SANITARY CLAMP TEE, 316L SS, 1-1/2" (YO-30560-40)
SANITARY CLAMP TEE, 304L SS, 1-1/2" (YO-30561-40).
More Details
Stainless Steel Hose Barb Fittings
Buy Stainless Steel Hose Barb Fittings Items Including:
Female Cam X 3/4-in Hose Barb Adapter, SS (QC-C075-SS)
Female Cam X 1-1/4-in Hose Barb Adapter, SS (QC-C125-SS)
Female Cam X 1-1/2-in Hose Barb Adapter, SS (QC-C150-SS)
Male Branch Tee, 316 SS, 10-32 UNF(M) x 1/16" hose barb (YO-31209-50).
More Details
Sanitary Fittings Hose Barb Adapters
Buy Sanitary Fittings Hose Barb Adapters Items Including:
Sanitary fittings, Plastic sanitary couplings, PVDF,1/4" ID, 1-9/16", (YO-31512-03)
Sani-Lock 1/2" Hose Barb Sanitary Adapter
Sani-Lock 3/4" Hose Barb Sanitary Adapter
Sani-Lock 1-1/2" Hose Barb Sanitary Adapter.
More Details
Sanitary to Hose Barb Fitting
Buy Sanitary to Hose Barb Fitting Items Including:
Sani-Lock 1/2" Hose Barb Sanitary Adapter
Sani-Lock 3/4" Hose Barb Sanitary Adapter
Sani-Lock 1-1/2" Hose Barb Sanitary Adapter
Barbed sanitary clamp adapter, 316L SS, 3/4" clamp x 1/8" tubing ID (YO-30560-08).
More Details
Hose Barb Clamps
Buy Hose Barb Clamps Items Including:
Sani-Lock 1/2" Hose Barb Sanitary Adapter
Sani-Lock 3/4" Hose Barb Sanitary Adapter
Sani-Lock 1-1/2" Hose Barb Sanitary Adapter
Adapter,1/2" Hose Barb (C105-14-645)
Adapter, NW 16 to 3/4" hose barb , 316L SS (C105-04-105).
More Details
Stainless Steel Hose Barb
Buy Stainless Steel Hose Barb Items Including:
Female Cam X 3/4-in Hose Barb Adapter, SS (QC-C075-SS)
Hose Barb 1/4"NPTm 304SS (YO-02930-35)
Male Branch Tee, 316 SS, 10-32 UNF(M) x 1/16" hose barb (YO-31209-50)
Male Branch Tee, 316 SS, 10-32 UNF(M) x 1/8" hose barb (YO-31209-51).
More Details
316l Stainless Steel Hose Barb Adapters
Buy 316l Stainless Steel Hose Barb Adapters Items Including:
Barbed sanitary clamp adapter, 316L SS, 1-1/2" clamp x 1-1/2" tubing I (YO-30560-14)
Sani-Lock 1/2" Hose Barb Sanitary Adapter
Sani-Lock 3/4" Hose Barb Sanitary Adapter
Sani-Lock 1" Hose Barb Sanitary Adapter.
More Details
Id Tube Clamp
Buy Id Tube Clamp Items Including:
Keck Tubing Adapter, 8 mm to 10 mm ID, 10/pk
Barbed sanitary clamp adapter, 316L SS, 3/4" clamp x 1/8" tubing ID (YO-30560-08)
Barbed sanitary clamp adapter, 316L SS, 3/4" clamp x 1/4" tubing ID (YO-30560-09).
More Details
Sanitary Fitting Flexible Hose
Buy Sanitary Fitting Flexible Hose Items Including:
Sanitary fittings, Plastic sanitary couplings, PVDF,1/4" ID, 1-9/16", (YO-31512-03)
Barbed sanitary clamp adapter, 316L SS, 1-1/2" clamp x 1-1/2" tubing I (YO-30560-14).
More Details
Sanitary Hose Barb 316l
Buy Sanitary Hose Barb 316l Items Including:
Barbed sanitary clamp adapter, 316L SS, 1-1/2" clamp x 1-1/2" tubing I (YO-30560-14)
Sani-Lock 1/2" Hose Barb Sanitary Adapter
Sani-Lock 3/4" Hose Barb Sanitary Adapter
Sani-Lock 1" Hose Barb Sanitary Adapter.
More Details
Hose Barb Tubing Clamp
Buy Hose Barb Tubing Clamp Items Including:
Barbed sanitary clamp adapter, 316L SS, 1-1/2" clamp x 1-1/2" tubing I (YO-30560-14)
BARBED SANITARY CLAMP ADAPTER, 304L SS, 1-1/2" CLAMP X 1-1/2" ID TUBIN (YO-30561-14).
More Details
316l Unpolished Stainless Tubing
Buy 316l Unpolished Stainless Tubing Items Including:
Cole-Parmer, Threaded Sanitary Clamp Adapter, 316L SS, 1.5" Clamp X 3/
Adapter, NW 16 to 1/4" NPT(M), 316L SS (C105-01-103)
Adapter, NW 16 to 1/8" NPT(), 316L SS (C105-01-106)
Adapter,1/4" NPT (F) X NW 25, 316L SS (C105-01-224).
More Details
Ss Tubing 0.012 Id
Buy Ss Tubing 0.012 Id Items Including:
Microbore PTFE Tubing, 0.012"ID x 0.030"OD, 100 ft/roll
Platinum-Cured SS Wire-Reinforced Silicone Hose 0.5 Inch ID
Platinum-Cured SS Wire-Reinforced Silicone Hose 0.75 Inch ID
Platinum-Cured SS Wire-Reinforced Silicone Hose 1.0 Inch ID.
More Details
Ss Tubing Adapter
Buy Ss Tubing Adapter Items Including:
Adapter, NW 16 to 1/4" Tube OD , 316L SS (C105-02-101)
Barbed sanitary clamp adapter, 316L SS, 3/4" clamp x 1/8" tubing ID (YO-30560-08)
Barbed sanitary clamp adapter, 316L SS, 1-1/2" clamp x 1-1/2" tubing I (YO-30560-14).
More Details
Sanitary Couplings Hose Assemblies
Buy Sanitary Couplings Hose Assemblies Items Including:
Plastic sanitary couplings; tubing ID, 1/4"; PP; 1 9/16" x 31/32" x 3 (YO-31512-00)
plastic sanitary couplings; tubing ID, 3/8"; PP; 1 11/16" x 31/32" x 5 (YO-31512-01).
More Details
414 Id Ss Tubing
Buy 414 Id Ss Tubing Items Including:
Platinum-Cured SS Wire-Reinforced Silicone Hose 0.75 Inch ID
Barbed sanitary clamp adapter, 316L SS, 3/4" clamp x 1/8" tubing ID (YO-30560-08)
Barbed sanitary clamp adapter, 316L SS, 3/4" clamp x 1/4" tubing ID (YO-30560-09).
More Details
Ss Tubing Clamps
Buy Ss Tubing Clamps Items Including:
Stainless Steel (SS) Hose Clamps, 11/16 x 1-1/4", box of 10
THREADED SANITARY CLAMP ADAPTER, 304L SS, 1" CLAMP X 1-1/2" NPT(M) (YO-30561-23)
316 SS Vacuum tubing, NW 16 flange, 10"
316 SS Vacuum tubing, NW 16 flange, 20".
More Details
Eccentric Tubing Reducers
Buy Eccentric Tubing Reducers Items Including:
ECCENTRIC SANITARY CLAMP REDUCER, 316L SS, 1-1/2" X 1" (YO-30560-05)
ECCENTRIC SANITARY CLAMP REDUCER, 316L SS, 2" X 1-1/2" (YO-30560-07)
ECCENTRIC SANITARY CLAMP REDUCER, 304L SS, 1-1/2" X 1" (YO-30561-05).
More Details
2 Steel Tubing Elbow
Buy 2 Steel Tubing Elbow Items Including:
Adapter,1/2" OD Tubing X NW 25 (C105-02-201)
BARBED SANITARY CLAMP ADAPTER, 304L SS, 2" CLAMP X 2" ID TUBING (YO-30561-15)
Coupling Body, 1/2" OD Tube (HFC13812)
Coupling Body, Panel Mount, Valved, 1/2" OD Tube (HFCD12812).
More Details
Stainless Tubing Elbow 375
Buy Stainless Tubing Elbow 375 Items Including:
SANITARY CLAMP ELBOW, 90°, 316L SS, 3/4" (YO-30560-30)
SANITARY CLAMP ELBOW, 90°, 316L SS, 1-1/2" (YO-30560-32)
SANITARY CLAMP ELBOW, 45°, 316L SS, 1-1/2" (YO-30560-36)
SANITARY CLAMP ELBOW, 90°, 304L SS, 1-1/2" (YO-30561-32).
More Details
Sanitary Tube Elbows
Buy Sanitary Tube Elbows Items Including:
SANITARY CLAMP ELBOW, 90°, 316L SS, 3/4" (YO-30560-30)
SANITARY CLAMP ELBOW, 90°, 316L SS, 1-1/2" (YO-30560-32)
SANITARY CLAMP ELBOW, 45°, 316L SS, 1-1/2" (YO-30560-36)
SANITARY CLAMP ELBOW, 90°, 304L SS, 1-1/2" (YO-30561-32).
More Details
Carbon Steel Tubing Elbows
Buy Carbon Steel Tubing Elbows Items Including:
SANITARY CLAMP ELBOW, 90°, 316L SS, 3/4" (YO-30560-30)
SANITARY CLAMP ELBOW, 90°, 316L SS, 1-1/2" (YO-30560-32)
SANITARY CLAMP ELBOW, 45°, 316L SS, 1-1/2" (YO-30560-36)
SANITARY CLAMP ELBOW, 90°, 304L SS, 1-1/2" (YO-30561-32).
More Details
70618
results
<<<Previous
20
21
22
23
24
25
26
27
28
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
36
37
38
40
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Steel Tube Couplings to Carbon Steel Tubing Elbows
>
31125329-s-toc_2 31125329stoc2