Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 26
Metric Hose Fittings
to
Cole Natural Polypropylene
Image
Product Detail
More Detail
Metric Hose Fittings
Buy Metric Hose Fittings Items Including:
Compression To Thread Adapter, Stainless Steel, 10mm OD Tubing X 1/2" (YO-31528-30).
More Details
To 6mm Tubing
Buy To 6mm Tubing Items Including:
Masterflex PTFE-tubing sets, 4mm ID, 6mm OD, set of two.
Barbed fittings, Straight Connectors, 6mm, Chemflour® PTFE, each (YO-06361-10)
Compression Reducing Union, Stainless Steel, 12mm X 6mm OD Tubing (YO-31528-41)
Glass Tubing, 6mm (YO-34742-06).
More Details
Stainless Compression Fittings for Stainless Hose
Buy Stainless Compression Fittings for Stainless Hose Items Including:
Compression To Thread Adapter, Stainless Steel, 10mm OD Tubing X 1/2" (YO-31528-30).
More Details
Npt to 1 4 Hose Barb
Buy Npt to 1 4 Hose Barb Items Including:
Barbed fittings, NPT male pipe adapter, Natural PP, 1/4" NPT x 1/4" I (YO-30610-38)
Barbed fittings, NPT male branch tees, Natural PP, 1/8" NPT x 1/4" ID, (YO-30610-72).
More Details
Pipe to Tube Adapter Fitting
Buy Pipe to Tube Adapter Fitting Items Including:
Male Pipe Adapter, Polypropylene, 1/8" x 5/32", 10 PACK (YO-30621-25)
Metal Quick-Disconnect NPT(F) Pipe Adapter Insert; Valved, 1/4" NPT(F) (MCD2604)
Compress to male threaded adapter, Pharmafluor PTFE, 1/16" OD x 1/8" N.
More Details
Adapters Hose to Threaded Male Pipe
Buy Adapters Hose to Threaded Male Pipe Items Including:
Barbed fittings, NPT male pipe adapter, Natural PP, 1/8" NPT x 1/4" ID (YO-30610-36)
Barbed fittings, NPT male pipe adapter, Natural PP, 1/8" NPT x 3/8" ID (YO-30610-37).
More Details
Adapter Straight Pipe to Tube
Buy Adapter Straight Pipe to Tube Items Including:
Barbed fittings, NPT male pipe adapter, Natural PP, 1/8" NPT x 1/4" ID (YO-30610-36)
Barbed fittings, NPT male pipe adapter, Natural PP, 1/8" NPT x 3/8" ID (YO-30610-37).
More Details
Npt Adapters Pipe to Hose
Buy Npt Adapters Pipe to Hose Items Including:
Barbed fittings, NPT male pipe adapter, Natural PP, 1/8" NPT x 1/4" ID (YO-30610-36)
Barbed fittings, NPT male pipe adapter, Natural PP, 1/8" NPT x 3/8" ID (YO-30610-37).
More Details
4 Natural Polypropylene Pipe
Buy 4 Natural Polypropylene Pipe Items Including:
Barbed fittings, NPT male pipe adapter, Natural PP, 1/8" NPT x 1/4" ID (YO-30610-36)
Barbed fittings, NPT male pipe adapter, Natural PP, 1/4" NPT x 3/8" ID (YO-30610-39).
More Details
Male Hose Fitting
Buy Male Hose Fitting Items Including:
Male Pipe Adapter, Polypropylene, 1/8" x 5/32", 10 PACK (YO-30621-25)
Hose barb to male threaded adapter, Pharmafluor™ PTFE, 1/4" NPT(M) (YO-06362-50)
Compress to male threaded adapter, Pharmafluor PTFE, 1/16" OD x 1/8" N.
More Details
Tube to 1 8 Pipe Fitting
Buy Tube to 1 8 Pipe Fitting Items Including:
Male Pipe Adapter, Polypropylene, 1/8" x 5/32", 10 PACK (YO-30621-25)
Compress to male threaded adapter, Pharmafluor PTFE, 1/16" OD x 1/8" N
Pipe fittings, Straight coupling, Chemfluor® PTFE, 1/8" NPT(F), 15 (YO-06469-16).
More Details
Tube Pipe to Tube Fittings
Buy Tube Pipe to Tube Fittings Items Including:
Barbed fittings, NPT male pipe adapter, Natural PP, 1/8" NPT x 1/4" ID (YO-30610-36)
Barbed fittings, NPT male pipe adapter, Natural PP, 1/8" NPT x 3/8" ID (YO-30610-37).
More Details
1.0 Male Tube Fittings
Buy 1.0 Male Tube Fittings Items Including:
Male Pipe Adapter, Polypropylene, 1/8" x 5/32", 10 PACK (YO-30621-25)
Male Pipe Adapter, Polypropylene, 10-32 UNF x 3/32", 25 PACK (YO-30621-01)
Male Pipe Adapter, Polypropylene, 10-32 UNF x 1/16", 25 PACK (YO-30621-04).
More Details
Tube to Pipe Thread
Buy Tube to Pipe Thread Items Including:
Barbed fittings, NPT male pipe adapter, Natural PP, 1/8" NPT x 1/4" ID (YO-30610-36)
Barbed fittings, NPT male pipe adapter, Natural PP, 1/8" NPT x 3/8" ID (YO-30610-37).
More Details
4 Hose or Piping
Buy 4 Hose or Piping Items Including:
Barbed fittings, NPT male pipe adapter, Natural PP, 1/8" NPT x 1/4" ID (YO-30610-36)
Barbed fittings, NPT male pipe adapter, Natural PP, 1/4" NPT x 3/8" ID (YO-30610-39).
More Details
Polypropylene Elbow Pipe Fittings
Buy Polypropylene Elbow Pipe Fittings Items Including:
Compression male pipe adapter elbow, 1/8" x 1/8", polypropylene, 10 Pa (YO-06386-05)
Compression male pipe adapter elbow, 1/4" x 1/8", polypropylene, 10 Pa (YO-06386-10).
More Details
Threaded Hose Adapter
Buy Threaded Hose Adapter Items Including:
Barbed fittings, NPT male pipe adapter elbows, Natural PP, 1/8" NPT x (YO-30610-54)
Barbed fittings, NPT male pipe adapter elbows, Natural PP, 1/8" NPT x (YO-30610-55).
More Details
Male Hose Elbow
Buy Male Hose Elbow Items Including:
Barbed fittings, NPT male pipe adapter elbows, Natural PP, 1/8" NPT x (YO-30610-54)
Barbed fittings, NPT male pipe adapter elbows, Natural PP, 1/8" NPT x (YO-30610-55).
More Details
Male to Barb Hose Adapter
Buy Male to Barb Hose Adapter Items Including:
Barbed fittings, NPT male pipe adapter elbows, Natural PP, 1/8" NPT x (YO-30610-54)
Barbed fittings, NPT male pipe adapter elbows, Natural PP, 1/8" NPT x (YO-30610-55).
More Details
Tubing to Male Pipe Thread
Buy Tubing to Male Pipe Thread Items Including:
Barbed fittings, NPT male pipe adapter, Natural PP, 1/8" NPT x 1/4" ID (YO-30610-36)
Barbed fittings, NPT male pipe adapter, Natural PP, 1/8" NPT x 3/8" ID (YO-30610-37).
More Details
Elbow Pipe Fitting
Buy Elbow Pipe Fitting Items Including:
Barbed fittings, NPT male pipe adapter elbows, Natural PP, 3/8" NPT x (YO-30610-59)
Barbed fittings, NPT male pipe adapter elbows, Natural PP, 1/8" NPT x (YO-30610-54).
More Details
Male to Male Hose Barb Connector
Buy Male to Male Hose Barb Connector Items Including:
Barbed fittings, NPT male branch tees, Natural PP, 1/8" NPT x 3/8" ID (YO-30610-73)
Male Branch Tee, 316 SS, 10-32 UNF(M) x 1/8" hose barb (YO-31209-51)
Pipe adapter elbow, Polypropylene, 1/8" x 5/16", 10 PACK (YO-30622-86).
More Details
Tube to Threaded Pipe Connectors
Buy Tube to Threaded Pipe Connectors Items Including:
Male pipe branch Tee, Polypropylene, 3/4" x 3/4", 10 PACKS (YO-30623-83)
Male pipe branch Tee, Polypropylene, 1/8" x 5/32", 10 PACKS (YO-30623-99)
Pipe adapter elbow, Polypropylene, 1/8" x 5/16", 100 PACK (YO-30626-86).
More Details
Pipe to Tubing Connectors
Buy Pipe to Tubing Connectors Items Including:
Male pipe branch Tee, Polypropylene, 3/4" x 3/4", 10 PACKS (YO-30623-83)
Male pipe branch Tee, Polypropylene, 1/8" x 5/32", 10 PACKS (YO-30623-99)
Pipe adapter elbow, Polypropylene, 1/8" x 5/16", 100 PACK (YO-30626-86).
More Details
Natural Polypropylene Fittings
Buy Natural Polypropylene Fittings Items Including:
Barbed fitting kits, Threaded elbow/tee kits, Natural PP, fittings kit (YO-41514-27)
Y-connectors, Polypropylene, 3/4", Pack Of 5 (Y0-12NP (5 PACK))
Barbed fittings, Straight Connector, Natural PP, 1/4" ID, 1/8", 1-31/3 (YO-30610-00).
More Details
Straight Hose Connectors
Buy Straight Hose Connectors Items Including:
Barbed fittings, Straight Connectors, 6mm, Chemflour® PTFE, each (YO-06361-10)
Straight compression union, Pharmafluor™ PTFE, 3mm tubing OD (YO-06396-12)
Barbed fittings, Straight Connector, Natural PP, 1/4" ID, 1/8", 1-31/3 (YO-30610-00).
More Details
Cole Hose Fittings
Buy Cole Hose Fittings Items Including:
Barbed fitting kits, Barbed connector/reducer kits, PP, small fittings (YO-31514-03)
Barbed fitting kits, Threaded barb adapter kits, Natural PP, small fit (YO-31514-11)
Bulkhead Fitting, PVDF, 3/8" NPT(F) Port X 1-13/16" OD (FK-TA03).
More Details
Cole-Parmer Natural Polypropylene Straight Barbed Connectors
Buy Cole-Parmer Natural Polypropylene Straight Barbed Connectors Items Including:
Straight barbed union, polypropylene, 3/16" ID, 10/pack (YO-30622-57)
Barbed fittings, Straight Connector, Natural PP, 1" ID, 25/32", 4", 1- (YO-30610-06)
Straight Connector, Polypropylene, 3/32", 100 PACK (YO-30626-46).
More Details
Hose Connector Tube
Buy Hose Connector Tube Items Including:
Barbed fittings, Straight Connectors, 6mm, Chemflour® PTFE, each (YO-06361-10)
Straight compression union, Pharmafluor™ PTFE, 3mm tubing OD (YO-06396-12)
Barbed fittings, Straight Connector, Natural PP, 1/4" ID, 1/8", 1-31/3 (YO-30610-00).
More Details
Cole Natural Polypropylene
Buy Cole Natural Polypropylene Items Including:
Barbed fittings, Straight Connector, Natural PP, 1" ID, 25/32", 4", 1- (YO-30610-06)
Barbed fittings, Reducing Connector, Natural PP,5/16" x 1/4" ID, 1/8", (YO-30610-07).
More Details
70618
results
<<<Previous
22
23
24
25
26
27
28
29
30
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
36
37
38
39
40
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Metric Hose Fittings to Cole Natural Polypropylene
>
31125329-s-toc_2 31125329stoc2