Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 27
Elbow Connector Silicone Hose
to
250 Inch Tubing Fittings
Image
Product Detail
More Detail
Elbow Connector Silicone Hose
Buy Elbow Connector Silicone Hose Items Including:
Barbed fittings, Elbow Connector, Natural PP,5/8" ID, 15/32", 1-29/32" (YO-30610-26)
Barbed fittings, Elbow Connector, Natural PP, 1" ID,25/32", 2-5/8", 2- (YO-30610-28).
More Details
Tubing Connector Kit
Buy Tubing Connector Kit Items Including:
Tubing Connector Kits; includes 10 tube end fittings with lock nuts, 1
Barbed fitting kits, Barbed elbow/tee/Y-connector kits, PP, small fitt (YO-31514-15)
Barbed fitting kits, Barbed connector/reducer kits, PP, small fittings (YO-31514-03).
More Details
Tube Fittings Kit
Buy Tube Fittings Kit Items Including:
Barbed fitting kits, Threaded elbow/tee kits, Natural PP, fittings kit (YO-31514-27)
Barbed fitting kits, Barbed connector/reducer kits, PP, small fittings (YO-31514-03)
Thermo Scientific Nalgene barbed bulkhead fitting kit, 1/2" tubing ID.
More Details
Tubing Fitting Tee Elbow
Buy Tubing Fitting Tee Elbow Items Including:
Barbed elbow/tee/Y-connector large fittings kits; Natural PP material (YO-31514-19)
Barbed fitting kits, Threaded elbow/tee kits, Natural PP, fittings kit (YO-31514-27)
Barbed fitting kits, BSPT, adapters/elbows/tees, Natural PP (YO-31514-30).
More Details
Inch Vacuum Tubing Fittings
Buy Inch Vacuum Tubing Fittings Items Including:
316 SS Vacuum tubing, NW 16 flange, 10"
316 SS Vacuum tubing, NW 16 flange, 20"
BARBED ELBOW, 1/4 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-72)
BARBED ELBOW, 3/8 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-73).
More Details
1&1 4 Hose Barb Tee Fitting
Buy 1&1 4 Hose Barb Tee Fitting Items Including:
Barbed fittings, NPT male tees, Natural PP, 1/4" NPT x 1/2" ID,1/4" (YO-30610-76)
Barbed fittings, NPT male tees, Natural PP, 1/2" NPT x 3/8" ID,1/4" (YO-30610-81).
More Details
Plastic Hose Fittings 1 4th Inch
Buy Plastic Hose Fittings 1 4th Inch Items Including:
BARBED ELBOW, 1/4 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-72)
BARBED ELBOW, 1/2 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-74)
Barbed to sanitary adapter, 1/4" ID to 1-1/2" sanitary clamp, animal-f (YO-31513-74).
More Details
Inch to 1 Inch Hose Barb
Buy Inch to 1 Inch Hose Barb Items Including:
BARBED ELBOW, 1/4 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-72)
BARBED ELBOW, 1/2 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-74)
BARBED ELBOW, 1/8 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-71).
More Details
Surgical Tubing 1 Inch
Buy Surgical Tubing 1 Inch Items Including:
BARBED ELBOW, 1/4 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-72)
BARBED ELBOW, 1/2 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-74)
BARBED ELBOW, 1/8 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-71).
More Details
Inch Aluminum Tubing Fittings
Buy Inch Aluminum Tubing Fittings Items Including:
Aluminum Tee Fittings, 63 ISO-K (C100-07-207)
Aluminum Tee Fittings, 63 ISO-K (C100-07-211)
BARBED ELBOW, 1/4 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-72)
BARBED ELBOW, 3/8 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-73).
More Details
Inch Copper Tube Fittings
Buy Inch Copper Tube Fittings Items Including:
BARBED ELBOW, 1/4 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-72)
BARBED ELBOW, 3/8 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-73)
BARBED ELBOW, 1/2 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-74).
More Details
1 Inch Ss Tubing
Buy 1 Inch Ss Tubing Items Including:
316 SS tube mixer; 1/4" OD; 21 elements (YO-04669-56)
316 SS tube mixer; 1/4" OD; 27 elements (YO-04669-58)
316 SS tube mixer; 1/4" OD; 34 elements (YO-04669-60)
Cole-Parmer, Barbed sanitary clamp adapter, 316L SS, 1-inch clamp x 1-.
More Details
Inch Poly Tubing Fittings
Buy Inch Poly Tubing Fittings Items Including:
Male luer integral lock ring to 1/8" semi-rigid tubing hose barb, poly (YO-45503-11)
BARBED ELBOW, 1/4 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-72)
BARBED ELBOW, 3/8 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-73).
More Details
Tubing Tee Fitting
Buy Tubing Tee Fitting Items Including:
Aluminum Tee Fittings, 63 ISO-K (C100-07-207)
Aluminum Tee Fittings, 63 ISO-K (C100-07-211)
Tee Fittings, 63 ISO-K, 316 SS (C100-07-207)
Tee Fittings, 100 ISO-K, 316 SS (C100-09-207)
Aluminum tee, NW 40 (C10516411)
More Details
2 Inch Hose
Buy 2 Inch Hose Items Including:
BARBED ELBOW, 1/2 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-74)
Barbed Tee, 1/2 Inch, Antimicrobial Polypropylene, Pack Of 5 (YO-06370-84)
Thermo Scientific Rejector with 2 inch (50.8 mm) OD for hose attacheme (YO-54502-31).
More Details
Inch Tubing Fittings
Buy Inch Tubing Fittings Items Including:
Barbed Fittings; Cross Connector, Clear PP, 1/4-inch Tubing ID, 50/pk (YO-31417-11)
BARBED ELBOW, 1/4 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-72)
BARBED ELBOW, 3/8 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-73).
More Details
8th Inch Tubing Fittings
Buy 8th Inch Tubing Fittings Items Including:
Barbed Fittings; Cross Connector, Clear PP, 1/4-inch Tubing ID, 50/pk (YO-31417-11)
BARBED ELBOW, 1/4 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-72)
BARBED ELBOW, 3/8 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-73).
More Details
Inch Plastic Tube Fittings
Buy Inch Plastic Tube Fittings Items Including:
Plastic luer fittings, PVC tubing with luer ends, 48" L, M/F fitting e (YO-30600-61)
BARBED ELBOW, 1/4 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-72)
BARBED ELBOW, 3/8 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-73).
More Details
1 Inch Corrug Ss Tubing
Buy 1 Inch Corrug Ss Tubing Items Including:
316 SS tube mixer; 1/4" OD; 21 elements (YO-04669-56)
316 SS tube mixer; 1/4" OD; 27 elements (YO-04669-58)
316 SS tube mixer; 1/4" OD; 34 elements (YO-04669-60)
Cole-Parmer, Barbed sanitary clamp adapter, 316L SS, 1-inch clamp x 1-.
More Details
1 Inch Square Tubing
Buy 1 Inch Square Tubing Items Including:
BARBED ELBOW, 1/4 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-72)
BARBED ELBOW, 1/2 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-74)
BARBED ELBOW, 1/8 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-71).
More Details
3 Inch Tubing
Buy 3 Inch Tubing Items Including:
BARBED ELBOW, 3/8 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-73)
BARBED TEE, 3/8 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-83)
BARBED ELBOW, 3/4 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-75).
More Details
4 Inch Pneumatic Hose
Buy 4 Inch Pneumatic Hose Items Including:
BARBED ELBOW, 1/4 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-72)
PendoTECH PressureMAT Single-Use Sensor, Polysulfone 1/4-inch hose bar (PREPS-N-025)
PendoTECH PressureMAT Single-Use Sensor, Polycarbonate 1/4-inch hose b (PRESS-N-025).
More Details
2 Inch Thin Wall Stainless Tubing
Buy 2 Inch Thin Wall Stainless Tubing Items Including:
Reducing bushing, 1/2" x 1/4", Stainless Steel, Each (YO-08539-93)
Reducing bushing, 1/2" x 3/8", Stainless Steel, Each (YO-08539-94)
Push-to-connect soft tube supports, 1/2" stem x 3/8" tube OD, 10/pack (YO-34007-39).
More Details
1 Inch Stainless Tubing for Boats
Buy 1 Inch Stainless Tubing for Boats Items Including:
Smart Gasket Clamp, 1/2" Tri-Clamp, 1 Port, Stainless Steel, Each
Smart Gasket Clamp, 1 1/2" Tri-Clamp, 1 Port, Stainless Steel, Each
Reducing bushing, 1/2" x 1/4", Stainless Steel, Each (YO-08539-93).
More Details
Hose Barbs Tee
Buy Hose Barbs Tee Items Including:
BARBED TEE, 3/8 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-83)
Male Branch Tee, 316 SS, 10-32 UNF(M) x 1/16" hose barb (YO-31209-50)
Barbed fittings, NPT male branch tees, Natural PP, 1/8" NPT x 1/4" ID, (YO-30610-72).
More Details
1.5 X1 5 Steel Tubing
Buy 1.5 X1 5 Steel Tubing Items Including:
Barbed fitting,Reducing Connector,1/2" x 5/8" Tube ID,PP (YO-06370-15)
Barbed Fittings; Straight Connector, 316 SS, 5/32" Tubing ID, 27/32" L (YO-31208-03)
Barbed Fittings; T Connector, 316 SS, 5/32" Tubing ID (YO-31208-33).
More Details
4 Inch Light Steel Tubing
Buy 4 Inch Light Steel Tubing Items Including:
316 Stainless Steel Tubing; dimensions, 3/16" ID, 1/4" OD, 1/32" wall; (YO-03300-10)
Pipe Mixer, 316 Stainless Steel, 1/4 inch NPT(M), 6 elements (YO-04669-05)
White Light Replacement Tube for Cole-Parmer 4-watt UV Lamp (YO-97620-58).
More Details
4 Inches Square Steel Tubing
Buy 4 Inches Square Steel Tubing Items Including:
Cole-Parmer, Barbed sanitary clamp adapter, 316L SS, 3/4-inch clamp x (YO-30700-58)
Cole-Parmer, Barbed sanitary clamp adapter, 316L SS, 3/4-inch clamp x (YO-30700-59)
BARBED ELBOW, 1/4 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-72).
More Details
4 Inch Round Stainless Tubing
Buy 4 Inch Round Stainless Tubing Items Including:
Cole-Parmer, Barbed sanitary clamp adapter, 316L SS, 3/4-inch clamp x (YO-30700-58)
Cole-Parmer, Barbed sanitary clamp adapter, 316L SS, 3/4-inch clamp x (YO-30700-59)
BARBED ELBOW, 1/4 inch, ANTIMICROBIAL POLYPROPYLENE, PACK OF 5 (YO-06370-72).
More Details
250 Inch Tubing Fittings
Buy 250 Inch Tubing Fittings Items Including:
Barbed Fittings; Cross Connector, Clear PP, 1/4-inch Tubing ID, 50/pk (YO-31417-11)
Reducing Connector, Polypropylene, 3/32" x 1/16", 250 PACK (YO-30625-95)
Reducing Connector, Polypropylene, 1/8" x 1/16", 250 PACK (YO-30626-23).
More Details
70618
results
<<<Previous
23
24
25
26
27
28
29
30
31
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
36
37
38
39
40
41
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Elbow Connector Silicone Hose to 250 Inch Tubing Fittings
>
31125329-s-toc_2 31125329stoc2