Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 30
Tee Fittings Npt Thread
to
Tubing Id(Inches) 0
Image
Product Detail
More Detail
Tee Fittings Npt Thread
Buy Tee Fittings Npt Thread Items Including:
Barbed fittings, NPT male tees, Natural PP, 1/4" NPT x 1/2" ID,1/4" (YO-30610-76)
Barbed fittings, NPT male tees, Natural PP, 1/2" NPT x 3/8" ID,1/4" (YO-30610-81).
More Details
Guaranteed I Tubing Fitting & Hose
Buy Guaranteed I Tubing Fitting & Hose Items Including:
Y-connectors, Polypropylene, 1/16", Pack Of 10 (YO-30726-41)
Y-connectors, Polypropylene, 3/32", Pack Of 10 (YO-30726-42)
Y-connectors, Polypropylene, 3/16", Pack Of 10 (YO-30726-44).
More Details
Guaranteed in Stock Program Tubing
Buy Guaranteed in Stock Program Tubing Items Including:
Pouch for Tubing Cutter 06438-90 (YO-06438-81)
Spring tubing clamp for up to 1/2"OD tubing, 3/bag (YO-06834-20)
Y-connectors, Polypropylene, 3/32", Pack Of 10 (YO-30726-42)
Y-connectors, Polypropylene, 1/8", Pack Of 10 (YO-30726-43).
More Details
Hose Barb Reducing Connectors
Buy Hose Barb Reducing Connectors Items Including:
Barbed fittings, Reducing Connector, Natural PP,5/16" x 1/4" ID, 1/8", (YO-30610-07)
Barbed fittings, Reducing Connector, Natural PP, 3/8" x 1/4" ID, 1/8", (YO-30610-08).
More Details
Reducing Hose Barb
Buy Reducing Hose Barb Items Including:
Barbed fittings, Reducing Connector, Natural PP,5/16" x 1/4" ID, 1/8", (YO-30610-07)
Barbed fittings, Reducing Connector, Natural PP, 3/8" x 1/4" ID, 1/8", (YO-30610-08).
More Details
1 Barbed Hose Connectors
Buy 1 Barbed Hose Connectors Items Including:
Male Branch Tee, 316 SS, 10-32 UNF(M) x 1/8" hose barb (YO-31209-51)
Aseptiquik Standard Insert, 1/2" Hose Barb, High Temperature
Snapquik Connector, Body, 1/8" Hose Barb, 25/pk
Snapquik Connector, Insert, 1/8" Hose Barb, 25/pk.
More Details
Tubing Id(Inches) 0.25 X 0.375
Buy Tubing Id(Inches) 0.25 X 0.375 Items Including:
Barbed fitting,Reducing Connector,1/4" x 3/8" Tube ID,PP (YO-06370-13)
Barbed fittings, Reducing Connector, Natural PP, 3/8" x 1/4" ID, 1/8", (YO-30610-08).
More Details
Cole-Parmer Natural Polypropylene Barbed Reducing Connectors
Buy Cole-Parmer Natural Polypropylene Barbed Reducing Connectors Items Including:
Barbed fittings, Reducing Connector, Natural PP,5/16" x 1/4" ID, 1/8", (YO-30610-07)
Barbed fittings, Reducing Connector, Natural PP, 3/8" x 1/4" ID, 1/8", (YO-30610-08).
More Details
Natural Polypropylene Barbed Reducing Connectors
Buy Natural Polypropylene Barbed Reducing Connectors Items Including:
Reducing Connector, Polypropylene, 5/32" x 1/8", 100 PACK (YO-30626-19)
Reducing Connector, Polypropylene, 1/8" x 1/16", 250 PACK (YO-30626-23)
Reducing Connector, Polypropylene, 1/4" x 5/32", 100 PACK (YO-30626-26).
More Details
Cole-Parmer Natural Polypropylene Barbed Y Connectors
Buy Cole-Parmer Natural Polypropylene Barbed Y Connectors Items Including:
Barbed fittings, Reducing Connector, Natural PP,5/16" x 1/4" ID, 1/8", (YO-30610-07)
Barbed fittings, Reducing Connector, Natural PP, 3/8" x 1/4" ID, 1/8", (YO-30610-08).
More Details
Tubing Id(Inches) 0.15625 X 0.125
Buy Tubing Id(Inches) 0.15625 X 0.125 Items Including:
Reducing Connector, Polypropylene, 5/32" x 1/8", 100 PACK (YO-30626-19).
More Details
Tubing Id(Inches) 0.25 X 0.15625
Buy Tubing Id(Inches) 0.25 X 0.15625 Items Including:
Reducing Connector, Polypropylene, 1/4" x 5/32", 100 PACK (YO-30626-26).
More Details
Tubing Id(Inches) 0.25 X 0.1875
Buy Tubing Id(Inches) 0.25 X 0.1875 Items Including:
Reducing Connector, Polypropylene, 1/4" x 3/16", 100 PACK (YO-30626-32).
More Details
Tubing Id(Inches) 0.3125 X 0.25
Buy Tubing Id(Inches) 0.3125 X 0.25 Items Including:
Barbed fittings, Reducing Connector, Natural PP,5/16" x 1/4" ID, 1/8", (YO-30610-07).
More Details
Tubing Id(Inches) 0.625 X 0.5
Buy Tubing Id(Inches) 0.625 X 0.5 Items Including:
Barbed fittings, Reducing Connector, Natural PP,5/8" x 1/2" ID, 11/32" (YO-30640-16).
More Details
Tubing Id(Inches) 0.625 X 0.25
Buy Tubing Id(Inches) 0.625 X 0.25 Items Including:
Barbed fittings, Reducing Connector, Natural PP, 5/8" x 1/4" ID, 1/8", (YO-30640-13).
More Details
Tubing Id(Inches) 0.375 X 0.3125
Buy Tubing Id(Inches) 0.375 X 0.3125 Items Including:
Barbed fittings, Reducing Connector, Natural PP, 3/8" x 5/16" ID, 3/16 (YO-30640-09).
More Details
Tubing Id(Inches) 0.625 X 0.3125
Buy Tubing Id(Inches) 0.625 X 0.3125 Items Including:
Barbed fittings, Reducing Connector, Natural PP, 5/8" x 5/16" ID, 3/16 (YO-30610-14).
More Details
Tubing Id(Inches) 0.75 X 0.625
Buy Tubing Id(Inches) 0.75 X 0.625 Items Including:
Barbed fittings, Reducing Connector, Natural PP, 3/4 x 5/8" ID, 15/32" (YO-30610-19).
More Details
Tubing Id(Inches) 0.625 X 0.375
Buy Tubing Id(Inches) 0.625 X 0.375 Items Including:
Barbed fittings, Reducing Connector, Natural PP, 5/8" x 3/8" ID, 1/4", (YO-30610-15).
More Details
Tubing Id(Inches) 0.75 X 0.375
Buy Tubing Id(Inches) 0.75 X 0.375 Items Including:
Barbed fittings, Reducing Connector, Natural PP, 3/4" x 3/8" ID, 1/4", (YO-30610-17).
More Details
Tubing Id(Inches) 0.5 X 0.3125
Buy Tubing Id(Inches) 0.5 X 0.3125 Items Including:
Barbed fitting,Straight connector,5/16" Tube ID,PP (YO-06370-05)
Barbed fittings, Reducing Connector, Natural PP,1/2" x 5/16" ID, 3/16 (YO-30610-11).
More Details
Bronze Hose to Hose Barb Fittings
Buy Bronze Hose to Hose Barb Fittings Items Including:
Barbed fitting kits, Barbed connector/reducer kits, PP, small fittings (YO-31514-03)
Barbed fitting kits, Threaded elbow/tee kits, Natural PP, fittings kit (YO-31514-27)
Temperature Sensor Cable for Hose Barb Fitting, 1/4" phone jack, 10'.
More Details
Barbed Tube Fittings
Buy Barbed Tube Fittings Items Including:
Thermo Scientific Nalgene barbed bulkhead fitting kit, 1/2" tubing ID
Thermo Scientific Nalgene barbed bulkhead fitting kit, 1/4" tubing ID
Barbed fittings, Reducing Connector, Clear PP,1/8" x 3/32" ID, 1/16", (YO-30506-07).
More Details
Natural Gas Aluminum Tubing Fittings
Buy Natural Gas Aluminum Tubing Fittings Items Including:
Barbed fitting kits, Threaded barb adapter kits, Natural PP, small fit (YO-31514-11)
Barbed fitting kits, Threaded elbow/tee kits, Natural PP, fittings kit (YO-31514-27)
Barbed fitting kits, BSPT, adapters/elbows/tees, Natural PP (YO-31514-30).
More Details
Natural Gas Tubing Fittings
Buy Natural Gas Tubing Fittings Items Including:
Barbed fitting kits, Threaded barb adapter kits, Natural PP, small fit (YO-31514-11)
Barbed fitting kits, Threaded barb adapter kits, Natural PP, large fit (YO-31514-35)
Barbed fitting kits, BSPT, adapters/elbows/tees, Natural PP (YO-31514-30).
More Details
Zinc Coated 2 Hose Barb Fittings
Buy Zinc Coated 2 Hose Barb Fittings Items Including:
Barbed fittings, Elbow Connector, Natural PP, 1" ID,25/32", 2-5/8", 2- (YO-30610-28)
Barbed fittings, T connector, Natural PP, 1/2" ID, 11/32", 2-21/32", 1 (YO-30610-32).
More Details
Tubing Id(Inches) 3/16
Buy Tubing Id(Inches) 3/16 Items Including:
Barbed fittings, Reducing Connector, Natural PP,1/2" x 5/16" ID, 3/16 (YO-30610-11)
Barbed fittings, Elbow Connector, Natural PP, 1/4" ID, 1/8", 1-3/16", (YO-30610-23)
Elbow, Polypropylene, 3/16", 10 PACK (YO-30622-75).
More Details
Bulk Barbed Tube Fittings
Buy Bulk Barbed Tube Fittings Items Including:
Thermo Scientific Nalgene barbed bulkhead fitting kit, 1/2" tubing ID
Thermo Scientific Nalgene barbed bulkhead fitting kit, 1/4" tubing ID
Barbed fittings, Reducing Connector, Clear PP,1/8" x 3/32" ID, 1/16", (YO-30506-07).
More Details
Tubing Id(Inches) 0.375
Buy Tubing Id(Inches) 0.375 Items Including:
Barbed fitting,Straight connector,3/8" Tube ID,PP (YO-06370-06)
Barbed fitting,Reducing Connector,1/4" x 3/8" Tube ID,PP (YO-06370-13)
Barbed fittings, Elbow Connector, Natural PP, 3/8" ID, 1/4", 1-7/16", (YO-30610-24).
More Details
70618
results
<<<Previous
26
27
28
29
30
31
32
33
34
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
10
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
36
37
38
39
40
41
42
43
44
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Tee Fittings Npt Thread to Tubing Id(Inches) 0
>
31125329-s-toc_2 31125329stoc2