Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 500
Compressed Air Pumps
to
General Purpose Spray Nozzles
Image
Product Detail
More Detail
Compressed Air Pumps
Buy Compressed Air Pumps Items Including:
Cole-Parmer Compressed Air Nozzle, Polyacetate, Flat Nozzle (600.130.56.BC.00.0)
Cole-Parmer Compressed Air Nozzle, Polyacetate, Flat Nozzle (600.332.56.BC.00.0).
More Details
Compressed Air Accessories
Buy Compressed Air Accessories Items Including:
Cole-Parmer Compressed Air Nozzle, ABS
Replacement Compressed-air filter; 6.0cfm; port size; 1/4" NPT(F); 5/b (5/050-05-BX)
Fixing/mounting Clamp For 21 cfm Filters (YO-29587-80)
Fixing/mounting Clamp For 42 And 64 cfm Filters (YO-29587-82).
More Details
Cole-Parmer Spray Nozzles
Buy Cole-Parmer Spray Nozzles Items Including:
316 stainless steel 90° full-cone spray nozzle, 0.053" orifice
PVDF 90° full-cone spray nozzle, 0.053" orifice
PVDF 90° full-cone spray nozzle, 0.074" orifice
Cole-Parmer Vaneless 90° Full-Cone Nozzle, 1/4" NPT(M) Inlet, 0.2.
More Details
Cole-Parmer Compressed Air Nozzles
Buy Cole-Parmer Compressed Air Nozzles Items Including:
Cole-Parmer Compressed Air Nozzle, Polyacetate, Flat Nozzle (600.130.56.BC.00.0)
Cole-Parmer Compressed Air Nozzle, Polyacetate, Flat Nozzle (600.332.56.BC.00.0).
More Details
Spray Pumps
Buy Spray Pumps Items Including:
316 stainless steel 90° full-cone spray nozzle, 0.053" orifice
316 stainless steel 90° full-cone spray nozzle, 0.059" orifice
Cole-Parmer Vaneless 90° Full-Cone Nozzle, 1/4" NPT(M) Inlet, 0.2
Cole-Parmer Compressed Air Nozzle, ABS.
More Details
Compressed Air Nozzles
Buy Compressed Air Nozzles Items Including:
Cole-Parmer Compressed Air Nozzle, Polyacetate, Flat Nozzle (600.130.56.BC.00.0)
Cole-Parmer Compressed Air Nozzle, Polyacetate, Flat Nozzle (600.332.56.BC.00.0).
More Details
Spray Nozzle
Buy Spray Nozzle Items Including:
316 stainless steel 90° full-cone spray nozzle, 0.053" orifice
Cole-Parmer Vaneless 90° Full-Cone Nozzle, 1/4" NPT(M) Inlet, 0.2
Cole-Parmer Compressed Air Nozzle, ABS
TBS SHUR/Freeze™ Cryogen spray with nozzle, 12 oz
can w/10 oz
More Details
Air Spray Nozzles
Buy Air Spray Nozzles Items Including:
Cole-Parmer Compressed Air Nozzle, Polyacetate, Flat Nozzle (600.130.56.BC.00.0)
Cole-Parmer Compressed Air Nozzle, Polyacetate, Flat Nozzle (600.332.56.BC.00.0).
More Details
Flat Spray Nozzle
Buy Flat Spray Nozzle Items Including:
Cole-Parmer Compressed Air Nozzle, Polyacetate, Flat Nozzle (600.130.56.BC.00.0).
More Details
Flat Nozzle
Buy Flat Nozzle Items Including:
Cole-Parmer Compressed Air Nozzle, Polyacetate, Flat Nozzle (600.130.56.BC.00.0).
More Details
Flat Spray Pumps
Buy Flat Spray Pumps Items Including:
Cole-Parmer Compressed Air Nozzle, Polyacetate, Flat Nozzle (600.130.56.BC.00.0).
More Details
Compression Pump
Buy Compression Pump Items Including:
Cole-Parmer Compressed Air Nozzle, Polyacetate, Flat Nozzle (600.130.56.BC.00.0)
Cole-Parmer Compressed Air Nozzle, Polyacetate, Flat Nozzle (600.332.56.BC.00.0).
More Details
Cole Accessories for Positive Displacement Pumps
Buy Cole Accessories for Positive Displacement Pumps Items Including:
Liquid level controller, 1/4" to 1" pumps, Normally closed, Close, PVC
Liquid level controller, 1-1/2" to 3" pumps, Normally closed, Close, S
Cole-Parmer Compressed Air Nozzle, Polyacetate, Flat Nozzle (600.130.56.BC.00.0).
More Details
Cole-Parmer Poly(Acetal)
Buy Cole-Parmer Poly(Acetal) Items Including:
Cole-Parmer Compressed Air Nozzle, Polyacetate, Flat Nozzle (600.130.56.BC.00.0).
More Details
Santoprene Diaphragm
Buy Santoprene Diaphragm Items Including:
Air-Powered Double-Diaphragm Pump, 1/4" Connections, PP / Santoprene w
High-Flow Double-Diaphragm Pump, 1/2" Connections, PP / Santoprene wet
Air-Powered Double-Diaphragm Pump, 1/4" Connections, Acetal / Santopre.
More Details
Pump Ad
Buy Pump Ad Items Including:
Hand-operated acid-transfer pump, 6 strokes/gallon, female buttress ad (236PF3P)
PTFE Pulse Dampeners With PTFE Diaphragm, 3/8" NPT
PTFE Pulse Dampeners With PTFE Diaphragm, 3/4" NPT
PTFE Pulse Dampeners With PTFE Diaphragm, 1" NPT.
More Details
Diaphragm Pumps Accessories
Buy Diaphragm Pumps Accessories Items Including:
Powder-Coated Base Plate for 75421-02, -12, -22, -72 Pumps
Surge Dampeners, 0.5" NPTF, PP Body, PTFE Diaphragm
Surge Dampeners, 0.75" NPTF, PP Body, PTFE Diaphragm
Surge Suppressors, 1" Flange, Aluminum Body, PTFE-Neoprene Diaphragm.
More Details
Positive Displacement Pipette Ptfe
Buy Positive Displacement Pipette Ptfe Items Including:
BRAND Positive Displacement Pipettors, 100-500 µL (2702904)
BRAND Positive Displacement Pipettors, 2-10 mL (2702912)
Brand Positive Displacement Pipettor Tips, PP, 100-500 µL, 10/pk
Brand Positive Displacement Pipettor Tips, PP, 200-1000 µL, 10/pk.
More Details
Accessories for Positive Displacement Pumps
Buy Accessories for Positive Displacement Pumps Items Including:
Liquid level controller, 1/4" to 1" pumps, Normally closed, Close, PVC
Liquid level controller, 1-1/2" to 3" pumps, Normally closed, Close, S
Pump, AODD Service Kit for SKU 75320-91
Pump, AODD Service Kit for SKU 75320-93.
More Details
Peristaltic Pump Pulse Dampener
Buy Peristaltic Pump Pulse Dampener Items Including:
PTFE Pulse Dampeners With PTFE Diaphragm, 3/8" NPT
PTFE Pulse Dampeners With PTFE Diaphragm, 3/4" NPT
Polypropylene Pulse Dampeners With Santoprene Diaphragm, 1-1/2" NPT
High-Flow Pulse Dampener for Series Q piston pump heads (PDHF).
More Details
Pulse Dampeners
Buy Pulse Dampeners Items Including:
PTFE Pulse Dampeners With PTFE Diaphragm, 3/8" NPT
PTFE Pulse Dampeners With PTFE Diaphragm, 3/4" NPT
Polypropylene Pulse Dampeners With Santoprene Diaphragm, 1-1/2" NPT
High-Flow Pulse Dampener for Series Q piston pump heads (PDHF).
More Details
Santoprene Accessories for Positive Displacement Pumps
Buy Santoprene Accessories for Positive Displacement Pumps Items Including:
Polypropylene Pulse Dampeners With Santoprene Diaphragm, 3/8" NPT
Polypropylene Pulse Dampeners With Santoprene Diaphragm, 1" NPT.
More Details
Ptfe Pulse Dampeners
Buy Ptfe Pulse Dampeners Items Including:
PTFE Pulse Dampeners With PTFE Diaphragm, 3/8" NPT
PTFE Pulse Dampeners With PTFE Diaphragm, 3/4" NPT.
More Details
Santoprene Pulse Dampeners
Buy Santoprene Pulse Dampeners Items Including:
Polypropylene Pulse Dampeners With Santoprene Diaphragm, 3/8" NPT
Polypropylene Pulse Dampeners With Santoprene Diaphragm, 1" NPT.
More Details
Cole-Parmer Multi-Purpose Spray Nozzles
Buy Cole-Parmer Multi-Purpose Spray Nozzles Items Including:
PVDF 90° full-cone spray nozzle, 0.053" orifice
PVDF 90° full-cone spray nozzle, 0.074" orifice.
More Details
Cole-Parmer General Purpose Spray Nozzles
Buy Cole-Parmer General Purpose Spray Nozzles Items Including:
PVDF 90° full-cone spray nozzle, 0.053" orifice
PVDF 90° full-cone spray nozzle, 0.074" orifice.
More Details
Multi-Purpose Spray Nozzles
Buy Multi-Purpose Spray Nozzles Items Including:
PVDF 90° full-cone spray nozzle, 0.053" orifice
PVDF 90° full-cone spray nozzle, 0.074" orifice
PVDF 90° full-cone spray nozzle, 0.106" orifice
Eutech Instruments replacement calibration set
One each of primary c (ECTN100CALKIT).
More Details
Cole-Parmer Pvdf Nozzles
Buy Cole-Parmer Pvdf Nozzles Items Including:
PVDF 90° full-cone spray nozzle, 0.053" orifice
PVDF 90° full-cone spray nozzle, 0.074" orifice
PVDF 90° full-cone spray nozzle, 0.106" orifice.
More Details
Cole Multi-Purpose Spray Nozzles
Buy Cole Multi-Purpose Spray Nozzles Items Including:
PVDF 90° full-cone spray nozzle, 0.053" orifice
PVDF 90° full-cone spray nozzle, 0.074" orifice.
More Details
General Purpose Spray Nozzles
Buy General Purpose Spray Nozzles Items Including:
PVDF 90° full-cone spray nozzle, 0.053" orifice
PVDF 90° full-cone spray nozzle, 0.074" orifice
PVDF 90° full-cone spray nozzle, 0.106" orifice
Eutech Instruments replacement calibration set
One each of primary c (ECTN100CALKIT).
More Details
70618
results
<<<Previous
496
497
498
499
500
501
502
503
504
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
100
200
300
400
410
420
430
440
450
460
470
480
485
486
487
488
489
490
491
492
493
494
495
496
497
498
499
500
501
502
503
504
505
506
507
508
509
510
511
512
513
514
520
530
540
550
560
570
580
590
600
700
800
900
1,000
1,100
1,200
1,300
1,400
2,000
2,354
Compressed Air Pumps to General Purpose Spray Nozzles
>
31125329-s-toc_2 31125329stoc2