Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 600
Guaranteed in Stock Program Centrifugal Pumps
to
Closed Impeller Pump
Image
Product Detail
More Detail
Guaranteed in Stock Program Centrifugal Pumps
Buy Guaranteed in Stock Program Centrifugal Pumps Items Including:
Centrifugal Magnetic Drive Pump with Enclosed Motor, Moderate-Flow Pol (YO-72010-00)
Centrifugal Magnetic Drive Pump with Enclosed Motor, Moderate-Flow Pol (YO-72010-10).
More Details
Motor Drives
Buy Motor Drives Items Including:
Basic Variable-Speed DC Motor Controller for 1/4 to 2 hp motors
Magnetic Drive Pump with 24 VDC Motor, Glass-Fiber Reinforced Polyprop (YO-72008-00)
Magnetic Drive Pump with 24 VDC Motor, Glass-Fiber Reinforced Polyprop (YO-72008-10).
More Details
Pump Moderate-Flow Centrifugal Pumps
Buy Pump Moderate-Flow Centrifugal Pumps Items Including:
304 SS Submersible Moderate-Flow Dewatering Pump, Automatic, 45 GPM, 1
Centrifugal Magnetic Drive Pump with Enclosed Motor, Moderate-Flow Pol (YO-72010-10)
Centrifugal Magnetic Drive Pump with Enclosed Motor, Moderate-Flow Pol (YO-72010-15).
More Details
Guaranteed I Centrifuges
Buy Guaranteed I Centrifuges Items Including:
Centrifugal Magnetic Drive Pump with Enclosed Motor, Moderate-Flow Pol (YO-72010-00)
Centrifugal Magnetic Drive Pump with Enclosed Motor, Moderate-Flow Pol (YO-72010-10)
9" Centrifuge Brush, 12/pk (610218)
Fixed-angle Rotor; 12 place x 15mL.
More Details
Ptfe Pumps - Centrifugal Pumps
Buy Ptfe Pumps - Centrifugal Pumps Items Including:
Centrifugal Magnetic Drive Pump with Enclosed Motor, Fluoroplastic, 3
(YO-72011-00)
Centrifugal Magnetic Drive Pump with Enclosed Motor, Fluoroplastic, 3
(YO-72011-05).
More Details
Ptfe Centrifugal Magnetic Drive Fluoroplastic Pumps
Buy Ptfe Centrifugal Magnetic Drive Fluoroplastic Pumps Items Including:
Centrifugal Magnetic Drive Pump with Enclosed Motor, Fluoroplastic, 3
(YO-72011-00)
Centrifugal Magnetic Drive Pump with Enclosed Motor, Fluoroplastic, 3
(YO-72011-05).
More Details
Ptfe Centrifugal Pumps
Buy Ptfe Centrifugal Pumps Items Including:
Centrifugal Magnetic Drive Pump with Enclosed Motor, Fluoroplastic, 3
(YO-72011-00)
Centrifugal Magnetic Drive Pump with Enclosed Motor, Fluoroplastic, 3
(YO-72011-05).
More Details
Magnetic Drive Pumps
Buy Magnetic Drive Pumps Items Including:
Magnetic Drive Pump with 24 VDC Motor, Glass-Fiber Reinforced Polyprop (YO-72008-00)
Magnetic Drive Pump with 24 VDC Motor, Glass-Fiber Reinforced Polyprop (YO-72008-10).
More Details
Centrifugal Mag Drive Pump
Buy Centrifugal Mag Drive Pump Items Including:
Centrifugal Magnetic Drive Pump with Enclosed Motor, Moderate-Flow Pol (YO-72010-00)
Centrifugal Magnetic Drive Pump with Enclosed Motor, Moderate-Flow Pol (YO-72010-05).
More Details
Centrifugal Magnetic Drive Fluoroplastic Pumps
Buy Centrifugal Magnetic Drive Fluoroplastic Pumps Items Including:
Centrifugal Magnetic Drive Pump with Enclosed Motor, Fluoroplastic, 3
(YO-72011-00)
Centrifugal Magnetic Drive Pump with Enclosed Motor, Fluoroplastic, 3
(YO-72011-05).
More Details
Ptfe Magnetic Drive Centrifugal Pumps
Buy Ptfe Magnetic Drive Centrifugal Pumps Items Including:
Centrifugal Magnetic Drive Pump with Enclosed Motor, Fluoroplastic, 3
(YO-72011-00)
Centrifugal Magnetic Drive Pump with Enclosed Motor, Fluoroplastic, 3
(YO-72011-05).
More Details
L/S Drive
Buy L/S Drive Items Including:
Magnetic Drive Pump with 24 VDC Motor, Glass-Fiber Reinforced Polyprop (YO-72008-00)
Magnetic Drive Pump with 24 VDC Motor, Glass-Fiber Reinforced Polyprop (YO-72008-10).
More Details
Noryl Pumps - Centrifugal Pumps
Buy Noryl Pumps - Centrifugal Pumps Items Including:
Noryl Corrosion-Resistant Mechanically Coupled Pump, 35 GPM, 115/230 V
Cole-Parmer Canned Motor Sealless Pump, Noryl Body, 3.5 GPM or 6 FT, 1 (YO-75561-03)
Cole-Parmer Canned Motor Sealless Pump, Noryl Body, 3.5 GPM or 6 FT, 1 (YO-75561-06).
More Details
Magnetic Sealless Pump
Buy Magnetic Sealless Pump Items Including:
Cole-Parmer Canned Motor Sealless Pump, 18 GPM, Noryl housing, 115 VAC
Cole-Parmer Canned Motor Sealless Pump, Noryl Body, 3.5 GPM or 6 FT, 1 (YO-75561-03)
Cole-Parmer Canned Motor Sealless Pump, Noryl Body, 3.5 GPM or 6 FT, 1 (YO-75561-06).
More Details
Canned Motor Sealless Pumps
Buy Canned Motor Sealless Pumps Items Including:
Cole-Parmer Canned Motor Sealless Pump, 18 GPM, Noryl housing, 115 VAC
Cole-Parmer Canned Motor Sealless Pump, Noryl Body, 3.5 GPM or 6 FT, 1 (YO-75561-03)
Cole-Parmer Canned Motor Sealless Pump, Noryl Body, 3.5 GPM or 6 FT, 1 (YO-75561-06).
More Details
Noryl Pumps
Buy Noryl Pumps Items Including:
Noryl Corrosion-Resistant Mechanically Coupled Pump, 35 GPM, 115/230 V
Cole-Parmer Canned Motor Sealless Pump, Noryl Body, 3.5 GPM or 6 FT, 1 (YO-75561-03)
Cole-Parmer Canned Motor Sealless Pump, Noryl Body, 3.5 GPM or 6 FT, 1 (YO-75561-06).
More Details
Noryl Centrifugal Pumps
Buy Noryl Centrifugal Pumps Items Including:
Noryl Corrosion-Resistant Mechanically Coupled Pump, 35 GPM, 115/230 V
Cole-Parmer Canned Motor Sealless Pump, Noryl Body, 3.5 GPM or 6 FT, 1 (YO-75561-03)
Cole-Parmer Canned Motor Sealless Pump, Noryl Body, 3.5 GPM or 6 FT, 1 (YO-75561-06).
More Details
Cole-Parmer Canned Motor Sealless Pumps
Buy Cole-Parmer Canned Motor Sealless Pumps Items Including:
Cole-Parmer Canned Motor Sealless Pump, 18 GPM, Noryl housing, 115 VAC.
More Details
Sealless Pumps
Buy Sealless Pumps Items Including:
Cole-Parmer Canned Motor Sealless Pump, 18 GPM, Noryl housing, 115 VAC
Cole-Parmer Canned Motor Sealless Pump, Noryl Body, 3.5 GPM or 6 FT, 1 (YO-75561-03)
Cole-Parmer Canned Motor Sealless Pump, Noryl Body, 3.5 GPM or 6 FT, 1 (YO-75561-06).
More Details
Motor and Pumps
Buy Motor and Pumps Items Including:
Cole-Parmer Canned Motor Sealless Pump, 18 GPM, Noryl housing, 115 VAC
Cole-Parmer Canned Motor Sealless Pump, Noryl Body, 3.5 GPM or 6 FT, 1 (YO-75561-03)
Cole-Parmer Canned Motor Sealless Pump, Noryl Body, 3.5 GPM or 6 FT, 1 (YO-75561-06).
More Details
Cole Canned Motor Sealless Pumps
Buy Cole Canned Motor Sealless Pumps Items Including:
Cole-Parmer Canned Motor Sealless Pump, 18 GPM, Noryl housing, 115 VAC.
More Details
Cole-Parmer Sealless Centrifugal Pumps
Buy Cole-Parmer Sealless Centrifugal Pumps Items Including:
Cole-Parmer Sealless Centrifugal Pump, Polypropylene Magnetic Drive, 3
Cole-Parmer Sealless Centrifugal Pump, Polypropylene Magnetic Drive, 6 (YO-72222-30)
Cole-Parmer Sealless Centrifugal Pump, Polypropylene Magnetic Drive, 6 (YO-72222-40).
More Details
High-Head Brushless Dc Magnetic Drive Pumps
Buy High-Head Brushless Dc Magnetic Drive Pumps Items Including:
Brushless DC Magnetic Drive Pump, High-Head Polypropylene, 1.6 GPM, 24
Brushless DC Magnetic Drive Pump, High-Head Polypropylene, 2.2GPM, 24 (YO-72025-15)
Brushless DC Magnetic Drive Pump, High-Head Polypropylene, 2.2GPM, 24 (YO-72025-25).
More Details
Centrifugal Magnetic Drive Polypropylene High-Head Pump
Buy Centrifugal Magnetic Drive Polypropylene High-Head Pump Items Including:
Centrifugal Magnetic Drive Pump with Enclosed Motor, High-Head Polypro (YO-72012-00)
Centrifugal Magnetic Drive Pump with Enclosed Motor, High-Head Polypro (YO-72012-05)
Brushless DC Magnetic Drive Pump, High-Head Polypropylene, 1.6 GPM, 24.
More Details
Magnetic Head Pumps
Buy Magnetic Head Pumps Items Including:
Centrifugal Magnetic Drive Pump with Enclosed Motor, High-Head Polypro (YO-72012-00)
Centrifugal Magnetic Drive Pump with Enclosed Motor, High-Head Polypro (YO-72012-05).
More Details
720 Drive Pump Heads
Buy 720 Drive Pump Heads Items Including:
Centrifugal Magnetic Drive Pump with Enclosed Motor, High-Head Polypro (YO-72012-00)
Centrifugal Magnetic Drive Pump with Enclosed Motor, High-Head Polypro (YO-72012-05)
Brushless DC Magnetic Drive Pump, High-Head Polypropylene, 1.6 GPM, 24.
More Details
Magnetic Impeller Pump
Buy Magnetic Impeller Pump Items Including:
Brushless DC Magnetic Drive Pump, PVDF, 3.1 GPM, 12 VDC, closed-impell
Brushless DC Magnetic Drive Pump, PVDF, 3.1 GPM, 24 VDC, closed-impell
Brushless DC Magnetic Drive Pump, PVDF, 3.6 GPM, 12 VDC, closed-impell.
More Details
Magnetic Drive Piston Pump
Buy Magnetic Drive Piston Pump Items Including:
Magnetic Drive Centrifugal Pump with Open Drip Proof (ODP) Motor, 5 GP (YO-07021-00)
Magnetic Drive Centrifugal Pump with Open Drip Proof (ODP) Motor, 5 GP (YO-07021-01).
More Details
Pvdf Couplings
Buy Pvdf Couplings Items Including:
Cole-Parmer Sealless Centrifugal Pump, Self-Priming PVDF Magnetic Driv (YO-72225-00)
PVDF Magnetic Drive Self-Priming Centrifugal Pumps, 78GPM - 40FT, thre
Cole-Parmer Sealless Centrifugal Pump, Self-Priming PVDF Magnetic Driv (YO-72225-10).
More Details
Closed Impeller Pump
Buy Closed Impeller Pump Items Including:
Closed-Impeller Close-Coupled Pump, 8 to 29 GPM, 115/230 VAC
Closed-Impeller Close-Coupled Pump, 19 to 47 GPM, 115/230 VAC
Closed-Impeller Close-Coupled Pump, 16 to 58 GPM, 115/230 VAC
Closed-Impeller Close-Coupled Pump, 16 to 58 GPM, 230/460 VAC.
More Details
70618
results
<<<Previous
596
597
598
599
600
601
602
603
604
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
100
200
300
400
500
510
520
530
540
550
560
570
580
585
586
587
588
589
590
591
592
593
594
595
596
597
598
599
600
601
602
603
604
605
606
607
608
609
610
611
612
613
614
620
630
640
650
660
670
680
690
700
800
900
1,000
1,100
1,200
1,300
1,400
1,500
2,000
2,354
Guaranteed in Stock Program Centrifugal Pumps to Closed Impeller Pump
>
31125329-s-toc_2 31125329stoc2