Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 70
1 Flexible Rubber Nylon Tubing
to
2 Id Neoprene Rubber Tubing
Image
Product Detail
More Detail
1 Flexible Rubber Nylon Tubing
Buy 1 Flexible Rubber Nylon Tubing Items Including:
Black Rubber Vacuum Tubing, 1/2" ID x 1 1/8" OD, 25ft
(YO-96143-12)
Black Rubber Vacuum Tubing, 1/2" ID x 1 1/8" OD, 25ft
(YO-96143-13)
Black Rubber Vacuum Tubing, 3/4" ID x 1 1/2" OD, 25ft
More Details
Od Tubing
Buy Od Tubing Items Including:
Steel tubing clamp for up to 3/8"OD tubing (YO-06404-10)
Steel tubing clamp for 3/4"OD tubing (YO-06404-30)
Adapter,1/2" OD Tubing X NW 25 (C105-02-201)
Compression Reducing Union, Stainless Steel, 12mm X 6mm OD Tubing (YO-31528-41).
More Details
R 2807 750 1 Black Tubing
Buy R 2807 750 1 Black Tubing Items Including:
Black Rubber Vacuum Tubing, 1/2" ID x 1 1/8" OD, 25ft
(YO-96143-12)
Black Rubber Vacuum Tubing, 1/2" ID x 1 1/8" OD, 25ft
(YO-96143-13)
Black Rubber Vacuum Tubing, 3/4" ID x 1 1/2" OD, 25ft
More Details
I P Tubing
Buy I P Tubing Items Including:
Black Rubber Vacuum Tubing, 3/16" ID x 9/16" OD, 25ft
(YO-96143-02)
Black Rubber Vacuum Tubing, 1/2" ID x 1 1/8" OD, 25ft
(YO-96143-12)
Black Rubber Vacuum Tubing, 5/8" ID x 1 3/8" OD, 25ft
More Details
Tubes for Tubing
Buy Tubes for Tubing Items Including:
Black Rubber Vacuum Tubing, 1/8" ID x 3/8" OD, 25ft
(YO-96143-01)
Black Rubber Vacuum Tubing, 3/16" ID x 9/16" OD, 25ft
(YO-96143-02)
Black Rubber Vacuum Tubing, 1/4" ID x 1/2" OD, 25ft
More Details
Tubing & Hose - all Types
Buy Tubing & Hose - all Types Items Including:
Male luer integral lock ring to 1/16" semi-rigid tubing hose barb, pol (YO-45503-07)
Black Rubber Vacuum Tubing, 3/16" ID x 9/16" OD, 25ft
(YO-96143-02)
Black Rubber Vacuum Tubing, 1/2" ID x 1 1/8" OD, 25ft
(YO-96143-12).
More Details
Vacuum Products
Buy Vacuum Products Items Including:
Vacuum Research Thermocouple Controller, 1 To 1000 Mtorr
Vacuum Research Thermocouple Sensor, 1/8" NPT (m)
Vacuum Research Thermocouple Sensor, 1/2" OD Flange
Black Rubber Vacuum Tubing, 3/16" ID x 9/16" OD, 25ft
(YO-96143-02).
More Details
Ss Tubing 1 4 Od
Buy Ss Tubing 1 4 Od Items Including:
Cole-Parmer Non-DEHP Lab Tubing 1/4" x 1/2", 50' pack
Cole-Parmer Non-DEHP Lab Tubing 1/4" x 3/8", 50' pack
Tygon High Purity Reinforced Tubing, 1/4"ID x 1/2"OD, 50 ft/pk
C-FLEX ULTRA TUBING 1/4" X 1/2", 50'.
More Details
Black Corrugated Pvc Flexible Tubing
Buy Black Corrugated Pvc Flexible Tubing Items Including:
Black Rubber Vacuum Tubing, 3/16" ID x 9/16" OD, 25ft
(YO-96143-02)
Black Rubber Vacuum Tubing, 1/2" ID x 1 1/8" OD, 25ft
(YO-96143-12)
Black Rubber Vacuum Tubing, 5/8" ID x 1 3/8" OD, 25ft
More Details
Flexible Tubing Large Od
Buy Flexible Tubing Large Od Items Including:
Black Rubber Vacuum Tubing, 3/16" ID x 9/16" OD, 25ft
(YO-96143-02)
Black Rubber Vacuum Tubing, 1/2" ID x 1 1/8" OD, 25ft
(YO-96143-12)
Black Rubber Vacuum Tubing, 5/8" ID x 1 3/8" OD, 25ft
More Details
Inch Od Flexible Tubing
Buy Inch Od Flexible Tubing Items Including:
Black Rubber Vacuum Tubing, 3/16" ID x 9/16" OD, 25ft
(YO-96143-02)
Black Rubber Vacuum Tubing, 1/2" ID x 1 1/8" OD, 25ft
(YO-96143-12)
Black Rubber Vacuum Tubing, 5/8" ID x 1 3/8" OD, 25ft
More Details
Weldable Flexible 1 4 Stainless Tubing
Buy Weldable Flexible 1 4 Stainless Tubing Items Including:
Cole-Parmer Non-DEHP Lab Tubing 1/4" x 1/2", 50' pack
Cole-Parmer Non-DEHP Lab Tubing 1/4" x 3/8", 50' pack
Cole-Parmer Non-DEHP Lab Tubing 1/4" x 7/16", 50' pack
Basket-tip filler nozzle, 1/4" ID, 10deg> tip.
More Details
Steel Tubing 1 4 Dia
Buy Steel Tubing 1 4 Dia Items Including:
304 Stainless Steel Tubing, 1/4" OD, 0.210" ID, 50ft roll (YO-34680-60)
Cole-Parmer Non-DEHP Lab Tubing 1/4" x 7/16", 50' pack
303/304 SS three-blade paddle assembly; 1/4" dia x 18"L, 2" paddle dia (YO-04449-07).
More Details
1 in Rubber Hose
Buy 1 in Rubber Hose Items Including:
Smooth FEP-lined Rubber Hose Assembly, 1-1/2" hex male, 10-ft length (YO-96556-14)
Smooth FEP-lined Convoluted Rubber Hose Assembly, 1" hex male, 6-ft le (YO-96557-02)
industrial Hose Pump, Natural Rubber Hose, 3.1 GPM, continuous duty, 1.
More Details
Product Type Tubing
Buy Product Type Tubing Items Including:
NMR tube probe for IQ150 handheld meter
PFA Test Tubes With Caps, 10 mL
PFA Test Tubes With Caps, 15 mL
PFA Test Tubes With Caps, 20 mL
PFA Test Tubes With Caps, 25 mL
More Details
Od X 0.007 Id Ss Tubing
Buy Od X 0.007 Id Ss Tubing Items Including:
316 SS tube mixer; 3/16" OD; 17 elements (YO-04669-50)
316 SS tube mixer; 3/16" OD; 21 elements (YO-04669-52)
316 SS tube mixer; 3/16" OD; 27 elements (YO-04669-54)
316 SS tube mixer; 1/4" OD; 21 elements (YO-04669-56).
More Details
Bunsen Burner Tubing
Buy Bunsen Burner Tubing Items Including:
Bunsen burner, adjustable with threaded needle valve, natural gas (H-5885)
Bunsen burner, adjustable with threaded needle valve, LP gas (H-5890)
Bunsen burner, Tirrill burner, LP gas (H-6225)
Bunsen burner, high-temperature burner, LP gas (H-5600).
More Details
Id Convoluted Rubber Tube
Buy Id Convoluted Rubber Tube Items Including:
Smooth FEP-lined Convoluted Rubber Hose Assembly, 1" hex male, 6-ft le (YO-96557-02)
Convoluted FEP tubing, 1/4"ID x 9/32"OD, 5 ft long (YO-06486-04)
Convoluted FEP tubing, 5/8"ID x 11/16"OD, 5 ft long (YO-06486-08).
More Details
Flexible Rubber Tubing
Buy Flexible Rubber Tubing Items Including:
Black Rubber Tubing, 3/16" ID x 5/16" OD, 25 ft (YO-96142-02)
Gum Rubber Tubing, 3/32" x 5/32", 25' per pack (057PXA-25')
Gum Rubber Tubing, 3/16" x 5/16", 25' per pack (099PXA-25').
More Details
Rubber Tubing 2.5 Mm
Buy Rubber Tubing 2.5 Mm Items Including:
Gum Rubber Tubing, 3/32" x 5/32", 25' per pack (057PXA-25')
Gum Rubber Tubing, 3/16" x 5/16", 25' per pack (099PXA-25')
Gum Rubber Tubing, 5/16" x 7/16", 25' per pack (138PXA-25').
More Details
Flexible Rubber Tubing Tulsa Ok
Buy Flexible Rubber Tubing Tulsa Ok Items Including:
Black Rubber Tubing, 3/16" ID x 5/16" OD, 25 ft (YO-96142-02)
Gum Rubber Tubing, 3/32" x 5/32", 25' per pack (057PXA-25')
Gum Rubber Tubing, 3/16" x 5/16", 25' per pack (099PXA-25').
More Details
Replacement Rubber Tubing for Hand Paddles
Buy Replacement Rubber Tubing for Hand Paddles Items Including:
Replacement pump tube, 39", CPVC
Black Rubber Tubing, 3/16" ID x 5/16" OD, 25 ft (YO-96142-02)
Gum Rubber Tubing, 3/32" x 5/32", 25' per pack (057PXA-25')
Gum Rubber Tubing, 1/8" x 1/4", 25' per pack (073PXA-25').
More Details
Stretch Rubber Tube Product
Buy Stretch Rubber Tube Product Items Including:
Black Rubber Tubing, 3/16" ID x 5/16" OD, 25 ft (YO-96142-02)
Gum Rubber Tubing, 3/32" x 5/32", 25' per pack (057PXA-25')
Gum Rubber Tubing, 3/16" x 5/16", 25' per pack (099PXA-25').
More Details
Flexible Solid Rubber Tubes
Buy Flexible Solid Rubber Tubes Items Including:
Black Rubber Tubing, 3/16" ID x 5/16" OD, 25 ft (YO-96142-02)
Gum Rubber Tubing, 3/32" x 5/32", 25' per pack (057PXA-25')
Gum Rubber Tubing, 3/16" x 5/16", 25' per pack (099PXA-25').
More Details
Cole Rubber Tubing
Buy Cole Rubber Tubing Items Including:
Black Rubber Tubing, 3/16" ID x 5/16" OD, 25 ft (YO-96142-02)
Gum Rubber Tubing, 3/32" x 5/32", 25' per pack (057PXA-25')
Gum Rubber Tubing, 3/16" x 5/16", 25' per pack (099PXA-25').
More Details
2 Inch Natural Rubber Tubing
Buy 2 Inch Natural Rubber Tubing Items Including:
Gum Rubber Tubing, 5/16" x 1/2", 25' per pack (140PXA-25')
Gum Rubber Tubing, 1/4x1/2
Black Rubber Vacuum Tubing, 1/4" ID x 1/2" OD, 25ft
(YO-96143-04)
Black Rubber Vacuum Tubing, 1/2" ID x 1" OD, 25ft
More Details
Mil R 6685 4 Rubber Tubing
Buy Mil R 6685 4 Rubber Tubing Items Including:
Black Rubber Tubing, 1/8" ID x 1/4" OD, 25 ft (YO-96142-01)
Black Rubber Tubing, 1/4 " ID x 3/8" OD, 25 ft (YO-96142-05)
Gum Rubber Tubing, 1/8x1/4
Black Rubber Vacuum Tubing, 1/4" ID x 1/2" OD, 25ft
(YO-96143-04).
More Details
Rubber Tubing 1 4 Inch Diameter
Buy Rubber Tubing 1 4 Inch Diameter Items Including:
Black Rubber Vacuum Tubing, 1/4" ID x 1/2" OD, 25ft
(YO-96143-04)
Black Rubber Vacuum Tubing, 1/4" ID x 1/2" OD, 25ft
(YO-96143-05)
Black Rubber Vacuum Tubing, 1/4" ID x 5/8" OD, 25ft
More Details
050 X 2 Inch Rubber Tubing
Buy 050 X 2 Inch Rubber Tubing Items Including:
Gum Rubber Tubing, 5/16" x 1/2", 25' per pack (140PXA-25')
Gum Rubber Tubing, 1/4x1/2
Black Rubber Vacuum Tubing, 1/4" ID x 1/2" OD, 25ft
(YO-96143-04)
Black Rubber Vacuum Tubing, 1/2" ID x 1" OD, 25ft
More Details
2 Id Neoprene Rubber Tubing
Buy 2 Id Neoprene Rubber Tubing Items Including:
Black Rubber Vacuum Tubing, 1/2" ID x 1" OD, 25ft
(YO-96143-10)
Black Rubber Vacuum Tubing, 1/2" ID x 1 1/8" OD, 25ft
(YO-96143-12)
Black Rubber Vacuum Tubing, 1/2" ID x 1 1/8" OD, 25ft
More Details
70618
results
<<<Previous
66
67
68
69
70
71
72
73
74
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
10
20
30
40
50
55
56
57
58
59
60
61
62
63
64
65
66
67
68
69
70
71
72
73
74
75
76
77
78
79
80
81
82
83
84
90
100
110
120
130
140
150
160
200
300
400
500
600
700
800
900
1,000
2,000
2,354
1 Flexible Rubber Nylon Tubing to 2 Id Neoprene Rubber Tubing
>
31125329-s-toc_2 31125329stoc2