Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 8
Tubing Id(Mm) 0
to
50 Amps
Image
Product Detail
More Detail
Tubing Id(Mm) 0.44
Buy Tubing Id(Mm) 0.44 Items Including:
3-Stop Tygon® Lab Tubing, 0.44 mm ID, 12/pack (CP95608-16)
2-Stop Tygon E-Lab Tubing, 0.44mm, 12/pack.
More Details
Tubing Id(Mm) 16
Buy Tubing Id(Mm) 16 Items Including:
3-Stop Tygon® Lab Tubing, 0.44 mm ID, 12/pack (CP95608-16)
2-Stop Tygon E-Lab Tubing, 0.44mm, 12/pack.
More Details
Small Electric Barrel Hose Pump
Buy Small Electric Barrel Hose Pump Items Including:
Remote Tap With Santoprene Seals & 5 Hose Complete
2-Stop Tygon® Lab Tubing, 0.19mm, 12/pack (CP95607-10)
2-Stop Tygon® Lab Tubing, 0.38mm, 12/pack (CP95607-14)
2-Stop Tygon® Lab Tubing, 0.51mm, 12/pack (CP95607-18).
More Details
Tygon Ismatec
Buy Tygon Ismatec Items Including:
3-Stop Tygon® Lab Tubing, 0.44 mm ID, 12/pack (CP95608-16)
3-Stop Tygon® Lab Tubing, 1.14 mm ID, 12/pack (CP95608-30)
3-Stop Tygon® Lab Tubing, 1.52 mm ID, 12/pack (CP95608-36)
3-Stop Tygon® Lab Tubing, 1.65 mm ID, 12/pack (CP95608-38).
More Details
Small Lab Gear Pump
Buy Small Lab Gear Pump Items Including:
Miniature gear pump, 0.67 GPM (2520 mL/min), 12 VDC lab model
3-Stop Tygon® Lab Tubing, 0.44 mm ID, 12/pack (CP95608-16)
3-Stop Tygon® Lab Tubing, 1.14 mm ID, 12/pack (CP95608-30)
3-Stop Tygon® Lab Tubing, 1.52 mm ID, 12/pack (CP95608-36).
More Details
Small Lab Pump
Buy Small Lab Pump Items Including:
3-Stop Tygon® Lab Tubing, 0.44 mm ID, 12/pack (CP95608-16)
3-Stop Tygon® Lab Tubing, 1.14 mm ID, 12/pack (CP95608-30)
3-Stop Tygon® Lab Tubing, 1.52 mm ID, 12/pack (CP95608-36)
3-Stop Tygon® Lab Tubing, 1.65 mm ID, 12/pack (CP95608-38).
More Details
Small Float Tube Pump
Buy Small Float Tube Pump Items Including:
Masterflex® C/L® Variable-Speed Tubing Pump; 1.7 to 10 rpm, 11 (YO-77122-00)
Masterflex® L/S® high-pressure pump, 115V
(YO-77914-00)
Tubing retainer kit for L/S high-performance pump head
2-Stop Tygon® Lab Tubing, 0.19mm, 12/pack (CP95607-10).
More Details
Ismatec Tubing, Fittings, and Hose
Buy Ismatec Tubing, Fittings, and Hose Items Including:
3-Stop Tygon® Lab Tubing, 0.44 mm ID, 12/pack (CP95608-16)
3-Stop Tygon® Lab Tubing, 1.14 mm ID, 12/pack (CP95608-30)
3-Stop Tygon® Lab Tubing, 1.52 mm ID, 12/pack (CP95608-36)
3-Stop Tygon® Lab Tubing, 1.65 mm ID, 12/pack (CP95608-38).
More Details
Small Lab Funnel Sets
Buy Small Lab Funnel Sets Items Including:
Small Lab Frame, Fiberglass
2-Stop Tygon® Lab Tubing, 0.19mm, 12/pack (CP95607-10)
2-Stop Tygon® Lab Tubing, 0.38mm, 12/pack (CP95607-14)
2-Stop Tygon® Lab Tubing, 0.51mm, 12/pack (CP95607-18)
2-Stop Tygon Lab Tubing, 1.85mm, 12/pack (CP95607-40).
More Details
Tubing Id(Mm) 0.38
Buy Tubing Id(Mm) 0.38 Items Including:
2-Stop Tygon® Lab Tubing, 0.38mm, 12/pack (CP95607-14)
PVC Solvent/Hydrocarbon Tubing, 0.38 mm ID, 12/pk (SC0287).
More Details
Flexible Tubing Plumbing Small Tubing
Buy Flexible Tubing Plumbing Small Tubing Items Including:
Tygon® pressure tubing, 1/2"ID x 3/4"OD, 50 ft/roll (YO-95634-02)
TYGON® Lab Tubing, 5/8"id X 13/16"OD, 50 Ft/pack
TYGON® Lab Tubing, 3/4"id X 1"OD, 50 Ft/pack
TYGON® Lab Tubing, 5/16"id X 1/2"OD, 50 Ft/pack.
More Details
Small Fittings for 1 4 Tubing
Buy Small Fittings for 1 4 Tubing Items Including:
Tygon® high-purity tubing, 1/8"ID x 1/4"OD, 50 ft/pack (YO-95635-01)
Male thread adapter, Click-N-Seal, 5/32" barb x 1/4"-28 UNF male threa (YO-96153-03)
TYGON® Lab Tubing, 1"id X 1-1/4"OD, 50 Ft/pack.
More Details
Small Tubing
Buy Small Tubing Items Including:
Small tube adapters, for 2 mL tubes, 2/pk
TYGON® Food and Beverage Tubing, 1/16"ID x 3/16"OD, 50 ft/pack
Tygon® pressure tubing, 1/2"ID x 3/4"OD, 50 ft/roll (YO-95634-02)
TYGON® Lab Tubing, 1"id X 1-1/4"OD, 50 Ft/pack.
More Details
Small Flexible Tubing
Buy Small Flexible Tubing Items Including:
Tygon® high-purity tubing, 1/8"ID x 1/4"OD, 50 ft/pack (YO-95635-01)
TYGON® Lab Tubing, 3/4"id X 1"OD, 50 Ft/pack
TYGON® Lab Tubing, 1"id X 1-1/4"OD, 50 Ft/pack
TYGON® Lab Tubing, 1/4"id X 1/2"OD, 50 Ft/pack.
More Details
Small Bore Tubing Fittings
Buy Small Bore Tubing Fittings Items Including:
2-Stop Tygon Lab Tubing, 1.85mm, 12/pack (CP95607-40)
2-Stop Tygon Lab Tubing, 2.29mm, 12/pack (CP95607-44)
Tygon® high-purity tubing, 1/16"ID x 3/16"OD, 50 ft/pack (YO-95635-00)
3-Stop Tygon® Lab Tubing, 0.44 mm ID, 12/pack (CP95608-16).
More Details
Small Diameter Tubing Fittings
Buy Small Diameter Tubing Fittings Items Including:
Dry Small Diameter Sampling Tube, 12" Length (PN007)
2-Stop Tygon® Lab Tubing, 0.19mm, 12/pack (CP95607-10)
2-Stop Tygon Lab Tubing, 1.85mm, 12/pack (CP95607-40)
Tygon® high-purity tubing, 1/16"ID x 3/16"OD, 50 ft/pack (YO-95635-00).
More Details
Small Rubber Tube Fittings
Buy Small Rubber Tube Fittings Items Including:
Atlas Fit® Rubber Coated Cotton Poly Glove Black and Gray Sm (320-S)
Atlas Fit® Rubber Coated Cotton Poly Glove Black and Gray Lg (320-L)
2-Stop Tygon® Lab Tubing, 0.19mm, 12/pack (CP95607-10)
2-Stop Tygon® Lab Tubing, 0.38mm, 12/pack (CP95607-14).
More Details
Tygon Tubing, Fittings, and Hose
Buy Tygon Tubing, Fittings, and Hose Items Including:
Masterflex® Tygon® tubing, nonphthlate/non-DEHP, 1.85mm ID, 1
3-Stop Tygon® Lab Tubing, 0.44 mm ID, 12/pack (CP95608-16)
3-Stop Tygon® Lab Tubing, 1.14 mm ID, 12/pack (CP95608-30)
3-Stop Tygon® Lab Tubing, 1.52 mm ID, 12/pack (CP95608-36).
More Details
Food Fittings for Tubing and Hose
Buy Food Fittings for Tubing and Hose Items Including:
TYGON® Food and Beverage Tubing, 1/16"ID x 3/16"OD, 50 ft/pack.
More Details
Small Tubing Products
Buy Small Tubing Products Items Including:
Masterflex® C/L® Variable-Speed Tubing Pump; 1.7 to 10 rpm, 11 (YO-77122-00)
Masterflex® C/L® Variable-Speed Tubing Pump; 13 to 80 rpm, 12 (YO-77122-12)
Tubing retainer kit for L/S high-performance pump head
Sensidyne® Gas Detection Tubes, Ethanol.
More Details
Food and Beverage Fittings
Buy Food and Beverage Fittings Items Including:
TYGON® Food and Beverage Tubing, 1/16"ID x 3/16"OD, 50 ft/pack
End cap, 3/4" (YO-30508-11).
More Details
Food Fittings
Buy Food Fittings Items Including:
Jumbo Refrigerator/freezer thermometer with a 5ml bottle that fits in (4627CP)
Jumbo Refrigerator/freezer thermometer with a 5ml bottle that fits in (4648CP)
TYGON® Food and Beverage Tubing, 1/16"ID x 3/16"OD, 50 ft/pack.
More Details
Small Lab Tube Furnace
Buy Small Lab Tube Furnace Items Including:
TYGON® Lab Tubing, 5/8"id X 13/16"OD, 50 Ft/pack
TYGON® Lab Tubing, 1"id X 1-1/4"OD, 50 Ft/pack
TYGON® Lab Tubing, 5/16"id X 1/2"OD, 50 Ft/pack
TYGON® Lab Tubing, 5/32"id X 7/32"OD, 50 Ft/pack.
More Details
Flexible Gas Tubing Fittings
Buy Flexible Gas Tubing Fittings Items Including:
Tygon® high-purity tubing, 1/16"ID x 3/16"OD, 50 ft/pack (YO-95635-00)
Tygon® high-purity tubing, 1/8"ID x 1/4"OD, 50 ft/pack (YO-95635-01)
Tygon® high-purity tubing, 5/16"ID x 7/16"OD, 50 ft/pack (YO-95635-04).
More Details
Tygon Vacuum Tubing
Buy Tygon Vacuum Tubing Items Including:
Tygon vacuum tubing, 3/16"ID x 9/16"OD, 10 ft/pack
Tygon vacuum tubing, 1/2"ID x 1-1/8"OD, 10 ft/pack
Tygon vacuum tubing, 5/8"ID x 1-3/8"OD, 10 ft/pack
Tygon vacuum tubing, 3/4"ID x 1-1/2"OD, 10 ft/pack.
More Details
Vacuum Hose Fittings
Buy Vacuum Hose Fittings Items Including:
Aluminum hose barb adapter, NW 16 to 10 mm (662511)
Adapter,1/2" Hose Barb (C105-14-645)
Adapter, SMOOTH, NW 16 to 1/2" hose barb, 316L SS (C105-04-104)
Tygon® vacuum tubing, 3/16"ID x 9/16"OD, 10 ft/pack (YO-06413-10).
More Details
Vacuum Tubing Fitting and Hose
Buy Vacuum Tubing Fitting and Hose Items Including:
Tygon® vacuum tubing, 3/16"ID x 9/16"OD, 10 ft/pack (YO-06413-10)
Aluminum hose barb adapter, NW 16 to 10 mm (662511)
Adapter,1/2" Hose Barb (C105-14-645)
Adapter, NW 16 to 3/4" hose barb , 316L SS (C105-04-105).
More Details
Vacuum Tubing Tubing, Fittings, and Hose
Buy Vacuum Tubing Tubing, Fittings, and Hose Items Including:
Adapter, SMOOTH, NW 16 to 1/2" hose barb, 316L SS (C105-04-104)
Tygon® vacuum tubing, 3/16"ID x 9/16"OD, 10 ft/pack (YO-06413-10)
Aluminum hose barb adapter, NW 16 to 10 mm (662511)
Adapter,1/2" Hose Barb (C105-14-645).
More Details
Flexible Hose Fittings
Buy Flexible Hose Fittings Items Including:
Tygon® pressure tubing, 1/2"ID x 3/4"OD, 50 ft/roll (YO-95634-02)
Tygon® high-purity tubing, 1/8"ID x 1/4"OD, 50 ft/pack (YO-95635-01)
Tygon S-50-HL, 1/32'' ID, 50' (YO-95665-00)
Tubing S-50-hl 3/32x5/32" 50' (YO-95665-03).
More Details
50 Amps
Buy 50 Amps Items Including:
Solid state relay, 50 amps, with 100 to 280 VAC control input
SSR 100-280 VAC Input AC Output 50 Amps 660V
CR4110S-50, 0 - 5 VDC output, AC True RMS
0-50 Amps
Fluke 50-Series II (Model 51) Thermocouple Thermometer with Single In (FLUKE-51-2 60HZ).
More Details
70618
results
<<<Previous
4
5
6
7
8
9
10
11
12
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Tubing Id(Mm) 0 to 50 Amps
>
31125329-s-toc_2 31125329stoc2