Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 900
Cable Mounting
to
Signet Ph Electrodes
Image
Product Detail
More Detail
Cable Mounting
Buy Cable Mounting Items Including:
Remote Optical Sensor w/8 ft Cable, Mounting Bracket, and 12" of Refle
Tie mounts for 7" to 11"L cable ties (08-462)
Cole-Parmer General Purpose pH Electrode, 10-ft Cable
Cole-Parmer High-pH Double-Junction pH Electrode, 10-ft Cable.
More Details
Double Junction Orp Probes
Buy Double Junction Orp Probes Items Including:
Cole-Parmer Removable Double-Junction ORP Electrode
Cole-Parmer Removable Double-Junction ORP Electrode, Tinned ends
Cole-Parmer Double-Junction ORP Electrode, Gold Sensor, 10-ft Cable
Cole-Parmer Removable Blue Glass Double-Junction pH Electrode.
More Details
Industrial Mounting Housing
Buy Industrial Mounting Housing Items Including:
Threaded In-Line Housing without Guard (YO-05993-76)
Threaded In-Line Housing with Kynar Guard
Threaded In-Line Housing with SS Guard (YO-05993-78)
Cole-Parmer Submersible Housing, CPVC
CPVC Submersible pH Electrode Holder, 100Ω RTD (YO-05725-42).
More Details
Gold Glasses
Buy Gold Glasses Items Including:
Cole-Parmer Single-Junction ORP Electrode, Gold Sensor, 10-ft Cable
Cole-Parmer Double-Junction ORP Electrode, Gold Sensor, 10-ft Cable.
More Details
Double Brand
Buy Double Brand Items Including:
Cole-Parmer Double-Junction ORP Electrode, Gold Sensor, 10-ft Cable
Cole-Parmer Double-Junction ORP Electrode, Platinum Sensor, 25-ft Cabl
Fluke BP291 Li-ion Battery pack, double capacity, for Fluke 190 Series.
More Details
Cole-Parmer 12-Mm High-Accuracy, Single-Junction, Ph Electrodes
Buy Cole-Parmer 12-Mm High-Accuracy, Single-Junction, Ph Electrodes Items Including:
Cole-Parmer 12-mm Glass body, High-Accuracy pH Electrode, 100 Ohm Pt R
Cole-Parmer High-Accuracy pH Electrode, 10-ft Cable
Cole-Parmer High-Accuracy pH Electrode, 100 Ohm PT RTD (Tinned-Ends),
Cole-Parmer High-Accuracy pH Electrode, 25-ft Cable.
More Details
High End Electrode Massager
Buy High End Electrode Massager Items Including:
Cole-Parmer Removable High pH Double-Junction pH Electrode, tinned-end.
More Details
High End Glass Vials
Buy High End Glass Vials Items Including:
Cole-Parmer Removable High pH Double-Junction pH Electrode, tinned-end.
More Details
High Precision Ph Meter
Buy High Precision Ph Meter Items Including:
Cole-Parmer High-Accuracy pH Electrode, 10-ft Cable
Cole-Parmer High-Accuracy pH Electrode, 100Ω Pt RTD, 10-ft Cable
Cole-Parmer High-Accuracy pH Electrode, 1 kΩ Pt RTD, 10-ft Cable
Cole-Parmer High-Accuracy pH Electrode, 100Ω Pt RTD, 25-ft Cable.
More Details
Body Ph Meter
Buy Body Ph Meter Items Including:
Thermo Scientific Orion Star LogR pH Meter with PerpHect glass-body pH
Cole-Parmer High-pH Double-Junction pH Electrode, 100 Ohm Pt RTD (Tinn (YO-05994-04)
Cole-Parmer General Purpose Double-Junction pH Electrode, 1 kΩ P (YO-05654-82).
More Details
Double Junction Ph Meter
Buy Double Junction Ph Meter Items Including:
Cole-Parmer General Purpose Double-Junction pH Electrode, 1 kΩ P (YO-05654-82)
Cole-Parmer High-pH Double-Junction pH Electrode, 100 Ohm Pt RTD (Tinn (YO-05994-04)
Cole-Parmer Removable Blue Glass Double-Junction pH Electrode, 100ome (YO-27009-11).
More Details
Double Junction Electrode
Buy Double Junction Electrode Items Including:
Cole-Parmer Removable High pH Double-Junction pH Electrode
Double junction electrode w/ Tuff-Tip (YO-27508-04)
Cole-Parmer Removable High pH Double-Junction pH Electrode, tinned-end
Cole-Parmer High-pH Double-Junction pH Electrode, 1 kΩ Pt RTD, 1.
More Details
Double Junction Calomel Electrode
Buy Double Junction Calomel Electrode Items Including:
Cole-Parmer High-pH Double-Junction pH Electrode, 100 Ohm Pt RTD (Tinn (YO-05994-04)
Cole-Parmer General Purpose Double-Junction pH Electrode, 1 kΩ P (YO-05654-82)
Cole-Parmer General Purpose Double-Junction pH Electrode, 1 kΩ P (YO-27013-82).
More Details
Double Junction Ph Electrode
Buy Double Junction Ph Electrode Items Including:
Cole-Parmer High-pH Double-Junction pH Electrode, 100 Ohm Pt RTD (Tinn (YO-05994-04)
Cole-Parmer General Purpose Double-Junction pH Electrode, 1 kΩ P (YO-05654-82)
Cole-Parmer General Purpose Double-Junction pH Electrode, 1 kΩ P (YO-27013-82).
More Details
High Temp Ph Meter
Buy High Temp Ph Meter Items Including:
YSI Ecosense pH Meter Only
YSI EcoSense pH Meter Kit
Cole-Parmer High-Accuracy pH Electrode, 10-ft Cable
Cole-Parmer 12-mm Glass body, High-Accuracy pH Electrode, 100 Ohm Pt R
Replacement high-performance pH electrode, combination, glass body, BN.
More Details
Double Junction Ph Spear
Buy Double Junction Ph Spear Items Including:
Cole-Parmer High-pH Double-Junction pH Electrode, 100 Ohm Pt RTD (Tinn (YO-05994-04)
Cole-Parmer General Purpose Double-Junction pH Electrode, 1 kΩ P (YO-05654-82)
Cole-Parmer Removable Blue Glass Double-Junction pH Electrode, 100ome (YO-27009-11).
More Details
Cole-Parmer High Ph Double-Junction Electrodes
Buy Cole-Parmer High Ph Double-Junction Electrodes Items Including:
Cole-Parmer High-pH Double-Junction pH Electrode, 1 kΩ Pt RTD, 1
Cole-Parmer High-pH Double-Junction pH Electrode, 10-ft Cable
Cole-Parmer High-pH Double-Junction pH Electrode, 25-ft Cable
Cole-Parmer Removable High pH Double-Junction pH Electrode.
More Details
Double Junction Reference Electrode
Buy Double Junction Reference Electrode Items Including:
Cole-Parmer High-pH Double-Junction pH Electrode, 100 Ohm Pt RTD (Tinn (YO-05994-04)
Cole-Parmer General Purpose Double-Junction pH Electrode, 1 kΩ P (YO-05654-82)
Cole-Parmer General Purpose Double-Junction pH Electrode, 1 kΩ P (YO-27013-82).
More Details
Double Junction
Buy Double Junction Items Including:
Cole-Parmer General Purpose Double-Junction pH Electrode, 1 kΩ P (YO-05654-82)
Cole-Parmer High-pH Double-Junction pH Electrode, 100 Ohm Pt RTD (Tinn (YO-05994-04)
Cole-Parmer Removable Blue Glass Double-Junction pH Electrode, 100ome (YO-27009-11).
More Details
Cole-Parmer General Purpose Single-Junction Ph Electrodes
Buy Cole-Parmer General Purpose Single-Junction Ph Electrodes Items Including:
Cole-Parmer Removable General-Purpose, Single-Junction pH Electrode
Cole-Parmer Removable General-Purpose, Single-Junction pH Electrode, T
Cole-Parmer General Purpose pH Electrode, 10-ft Cable
Cole-Parmer General Purpose pH Electrode, 25-ft Cable.
More Details
Cole-Parmer General Purpose Housings
Buy Cole-Parmer General Purpose Housings Items Including:
Cole-Parmer General Purpose pH Electrode, 1 kΩ Pt RTD, 10-ft Cab
Cole-Parmer General Purpose pH Electrode, 10-ft Cable
Cole-Parmer General Purpose pH Electrode, 25-ft Cable
Cole-Parmer General Purpose Double-Junction pH Electrode, 25-ft Cable.
More Details
General Purpose Housings
Buy General Purpose Housings Items Including:
Cole-Parmer General Purpose pH Electrode, 10-ft Cable
Cole-Parmer 12-mm Glass Body, General Purpose, Double-Junction pH Ele
Cole-Parmer 12-mm Glass Body, General Purpose, Double-Junction pH Ele
Cole-Parmer General Purpose pH Electrode, 25-ft Cable.
More Details
Cole General Purpose M
Buy Cole General Purpose M Items Including:
Cole-Parmer General Purpose pH Electrode, 10-ft Cable
Cole-Parmer 12-mm Glass Body, General Purpose, Double-Junction pH Ele
Cole-Parmer 12-mm Glass Body, General Purpose, Double-Junction pH Ele
Cole-Parmer General Purpose pH Electrode, 25-ft Cable.
More Details
Low Cost Ph Meter
Buy Low Cost Ph Meter Items Including:
Cole-Parmer® Low-Cost Loop-Powered Meter, NEMA 4X, Backlit (YO-94799-02)
Low-Cost Specific Gravity Meter, 0.750 to 1.000 SGU, °C model (DSG-11-075100-C-B)
Tintometer Comparator 2000, AF328 Hazen/Alpha Low
Oakton EcoTestr TDS Low pocket TDS tester.
More Details
Conductivity Meter Low Volume
Buy Conductivity Meter Low Volume Items Including:
100 Ω Pt RTD Conductivity Cell for ultra-low range measurements, (YO-19500-02)
100 Ω Pt RTD Conductivity Cell for ultra-low range measurements, (YO-19500-04)
pH Electrode, low conductivity, bulb, 3KOhm Balco thermistor, 3/4" NPT.
More Details
Signet 2724 Ph Electrode
Buy Signet 2724 Ph Electrode Items Including:
pH Electrode, flat surface, Pt-1000 ATC, 3/4" NPT threads
pH Electrode, flat surface, Pt-1000 ATC, ISO 7/1 R3/4 threads
pH Electrode, bulb, HF resistant, Pt-1000 ATC, ISO 7/1 R3/4 threads
pH Electrode, bulb, Pt-1000 ATC, 3/4" NPT threads.
More Details
Hf-Resistant Electrode
Buy Hf-Resistant Electrode Items Including:
Cole-Parmer HF Resistant Submersible pH Electrode, CPVC, 100Ω RT (YO-27108-02)
Cole-Parmer HF Resistant Submersible pH Electrode, CPVC, 1 kΩ RT (YO-27108-04)
pH Electrode, bulb, HF resistant, 3KOhm Balco thermistor, 3/4" NPT thr.
More Details
Low Conductance in Water
Buy Low Conductance in Water Items Including:
pH Electrode, low conductivity, bulb, 3KOhm Balco thermistor, 3/4" NPT
100 Ω Pt RTD Conductivity Cell for ultra-low range measurements, (YO-19500-02)
100 Ω Pt RTD Conductivity Cell for ultra-low range measurements, (YO-19500-04).
More Details
Balco Thermistor
Buy Balco Thermistor Items Including:
pH Electrode, flat surface, 3KOhm Balco thermistor ATC, 3/4" NPT threa
pH Electrode, bulb, HF resistant, 3KOhm Balco thermistor, 3/4" NPT thr
pH Electrode, low conductivity, bulb, 3KOhm Balco thermistor, 3/4" NPT.
More Details
Signet Ph Electrodes
Buy Signet Ph Electrodes Items Including:
pH Electrode, flat surface, Pt-1000 ATC, 3/4" NPT threads
pH Electrode, flat surface, Pt-1000 ATC, ISO 7/1 R3/4 threads
pH Electrode, bulb, HF resistant, Pt-1000 ATC, ISO 7/1 R3/4 threads
pH Electrode, bulb, Pt-1000 ATC, 3/4" NPT threads.
More Details
70618
results
<<<Previous
896
897
898
899
900
901
902
903
904
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
100
200
300
400
500
600
700
800
810
820
830
840
850
860
870
880
885
886
887
888
889
890
891
892
893
894
895
896
897
898
899
900
901
902
903
904
905
906
907
908
909
910
911
912
913
914
920
930
940
950
960
970
980
990
1,000
1,100
1,200
1,300
1,400
1,500
1,600
1,700
1,800
2,000
2,354
Cable Mounting to Signet Ph Electrodes
>
31125329-s-toc_2 31125329stoc2