Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 1
Labpette Liquid Handling
to
Small Enclosures
Image
Product Detail
More Detail
Labpette Liquid Handling
Buy Labpette Liquid Handling Items Including:
Labpette Discovery Pipette 0.5 - 10.0 uL (P3940-10)
Labpette Discovery Pipette 10 - 100 uL (P3940-100)
Labpette Discovery Pipette 20 - 200 uL (P3940-200)
Labpette Discovery Pipette 100- 1000 uL (P3940-1000)
Labpette Discovery Pipette 0.1 - 2.0 uL.
More Details
Stainless Steel Manifold
Buy Stainless Steel Manifold Items Including:
304 Stainless Steel manifold; 2 outlets; 1/4" inlet NPT(F) (YO-31521-31)
304 Stainless Steel manifold; 2 outlets; 3/8" inlet NPT(F) (YO-31521-33)
304 Stainless Steel manifold; 2 outlets; 1/2" inlet NPT(F) (YO-31521-35).
More Details
Data Logger
Buy Data Logger Items Including:
MicroLite II Temperature USB Data Logger with 8k Sample Memory
MicroLite II Current USB Data Logger
MicroLite II Voltage USB Data Logger
MicroLite II External NTC Temperature USB Data Logger
Track It™ data logger USB interface cable (YO-30003-04).
More Details
Chart Recorder Charts
Buy Chart Recorder Charts Items Including:
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C304)
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C312)
Replacement Chart Paper for Super-Compact Temperature Chart Recorders, (C320).
More Details
Cole-Parmer Noncontact Tachometers
Buy Cole-Parmer Noncontact Tachometers Items Including:
Convertible Laser Tachometer, LCD model (DT-205LR)
Pocket laser tachometer (PLT-5000)
1/8 DIN panel-mount standard single-pulse tachometer, 115 VAC (YO-08212-20)
1/8 DIN panel-mount programmable tachometer/counter/totalizer, 230 VAC (YO-08212-45).
More Details
Mixers Heavy Duty
Buy Mixers Heavy Duty Items Including:
Heavy duty, gear-drive explosion proof mixer; 1/2-hp, 36"L shaft, 8" d (YO-50311-42)
Heavy duty, gear-drive explosion proof mixer; 1-hp, 60"L shaft, 14" im (YO-50311-52)
Heavy-duty, reversible mixer, 115 VAC (YO-50402-30).
More Details
Cole-Parmer Bulkhead Connectors
Buy Cole-Parmer Bulkhead Connectors Items Including:
Bulkhead connectors, nylon, 1/8" NPT(F) x 5/8" OD, pack of 6 (YO-06349-10)
Pipe fittings, Swivel bulkhead connector, PVC, 3" NPT (F),3-1/4", 8-3/ (YO-31800-20)
Self Aligning Swivel Bulkhead fitting, PVC, 1" (YO-06225-80).
More Details
15 Gallon Plastic Drum
Buy 15 Gallon Plastic Drum Items Including:
Cone bottom tank, 15 gallons, 45° cone (YO-06960-26)
3" wide flexible drum heater, 15 gallon, 500 watt, 120 VAC, 70-425deg (YO-09200-10)
Drip Sentinel Drum-top Workstation For 55 Gallon Steel Drums
Justrite Drum Lock, Plastic Drum Set (08509).
More Details
Sodium Sulfite Anhydrous
Buy Sodium Sulfite Anhydrous Items Including:
SODIUM SULFITE ANHYDROUS 25G (YO-88234-56).
More Details
250 Ml Erlenmeyer Flask
Buy 250 Ml Erlenmeyer Flask Items Including:
Corning 250 mL PC Flask w/vented cap, 50/CS
Thermo Scientific Low Cost Orbital Shaker for 250 mL Erlenmeyer flasks (2346Q)
Thermo Scientific Low Cost Orbital Shaker for 250 mL Erlenmeyer flasks (2346-1CEQ)
PYREX Brand 4995 flask; 250 mL, case of 6.
More Details
Latex Free Exam Gloves
Buy Latex Free Exam Gloves Items Including:
Best N-DEX Nitrile Exam Gloves, Powder-Free, medium, 100/pack
Best N-DEX Nitrile Exam Gloves, Powder-Free, large, 100/pack
Best N-DEX Nitrile Exam Gloves, Powder-Free, extra-large, 100/pack
AURELIA™ Velocity™ Powder-Free Latex Gloves; size, medium.
More Details
Industrial Transducers
Buy Industrial Transducers Items Including:
Heavy- Duty Industrial Pressure Transmitter, 30" Hg to 0 psi; 4 to 20
Heavy- Duty Industrial Pressure Transmitter, 30" Hg to 30 psi; 4 to 20
Heavy- Duty Industrial Pressure Transmitter, 0 to 1000 psi; 1 to 5 VDC.
More Details
High Pressure Metering Pump
Buy High Pressure Metering Pump Items Including:
Pump, Cole-Parmer Deluxe Peristaltic Metering Pump, Max flow of 33.3 G
Economical High-Pressure Single-Piston Pump; 8.0 mL/min Max Flow, 230 (B-100-S-2-CE (#1442))
Pumps, Metering Pumps Replacement Tube Set for Mid-Flow Range Cole-Par (YO-74204-01).
More Details
Compact M
Buy Compact M Items Including:
Ismatec™ compact analog pump, 2 channels, 1.6 to 160 rpm, 0.004 to (YO-78016-10)
Ismatec™ compact digital pump, 2 channels, 1.6 to 160 rpm, 0.004 to (YO-78016-45)
Ismatec™ compact digital pump, 2 channels, 1.6 to 160 rpm, 0.004 to (YO-78017-00).
More Details
40 Chest
Buy 40 Chest Items Including:
Thermo Scientific Revco CxF -40°C Chest Freezer, 12.7 cubic feet, (ULT1350-10-D)
Thermo Scientific Revco CxF -40°C Chest Freezer, 12.7 cubic feet, (ULT1350-10-V)
Thermo Scientific Revco CxF -40°C Chest Freezer, 12.7 cubic feet, (ULT1350-10-A).
More Details
Thermocouple Gas
Buy Thermocouple Gas Items Including:
Type T Air/Gas Thermocouple Probe; 8"L (112601B)
Type J Air/Gas Thermocouple Probe; 8"L (112601B)
Type J Low-cost air/gas thermocouple probe; 4.5"L (070197A-J)
Type K Low-cost air/gas thermocouple probe; 4.5"L (070197A-K).
More Details
Medium Pack
Buy Medium Pack Items Including:
Advantec Grade No
2 Qualitative Filter Paper; 5.5 cm dia.; pack of 10
Advantec Grade No
2 Qualitative Filter Paper; 7.0 cm dia.; pack of 10
Advantec Grade No
More Details
Varipet Dispensers
Buy Varipet Dispensers Items Including:
Varipet repetitive syringe dispensers; volume range, 0.0 - 1.0 mL; inc.
More Details
Monitor 70
Buy Monitor 70 Items Including:
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Digital (YO-08895-70)
Air Flow Monitor for Bio Safety Cabinets, 30 to 400 FPM Monitoring Ran
Primary Formaldehyde Filters for 30" Self-Monitoring Ductless Fume Hoo (YO-33713-70).
More Details
Ptfe Pumps - Positive Displacement Pumps
Buy Ptfe Pumps - Positive Displacement Pumps Items Including:
PTFE flexible liner pump, Nordel rubber liner, 1 GPM, 115/230 VAC
PTFE flexible liner pump, Nordel rubber liner, 3 GPM, 115/230 VAC
Surge Dampeners, 0.5" NPTF, PP Body, PTFE Diaphragm
Surge Dampeners, 0.5" NPTF, PVDF Body, PTFE Diaphragm.
More Details
Scienceware Spill Cleanup
Buy Scienceware Spill Cleanup Items Including:
Mercury collector jar (F36250).
More Details
Dew Point Instruments
Buy Dew Point Instruments Items Including:
Digital Thermohygrometer Pen With Dew Point And Heat Index (SAM800IND)
Digital Psychrometer, RH, Temp, & Dew Point (LAM880D).
More Details
Bacharach Combustion
Buy Bacharach Combustion Items Including:
Fyrite Tech Combustion Analyzer
Bacharach PCA 3 Combustion Analyzer, with O2, and CO sensors
Combustible sensor
Bacharach PCA 3 Combustion Analyzer, with O2, CO, and NO sensors
Bacharach PCA 3 Combustion Analyzer, with O2, CO, and CO (high range).
More Details
Coating Gauges
Buy Coating Gauges Items Including:
Dftf:Dgtl Fe Coating Gauge Thickness Gauge
Defelsko PosiTector 6000FS1 Std Thickness coating gauge for non-magnet
Defelsko PosiTector 6000FN1 Std Thickness coating gauge for non-magnet
W/Built-In Probe Basic Non-Magnetic Coatings On Ferrous Metals.
More Details
Thermo Scientific Forma
Buy Thermo Scientific Forma Items Including:
Thermo Scientific MaxQ 8000 Stackable Incubated Shaker, 120V.
More Details
Seripettor General Accessories
Buy Seripettor General Accessories Items Including:
Replacement Cartridges for BrandTech Seripettor Economy Dispenser, 1.0
Replacement Cartridges for BrandTech Seripettor Economy Dispenser, 2.5.
More Details
Probe High Voltage
Buy Probe High Voltage Items Including:
Bartol Mag-Probe High Sensitivity Noncontact Probe
Tektronix High Voltage Single-ended Probe, 100X, 250 MHz (P5100)
Tektronix High Voltage Single-ended Probe, 1000X, 75 MHz, 10 FT (P6015A)
Tektronix TPP0850 High Voltage Probe; 50X, 800 MHZ, 2.5 kV.
More Details
800 Amps
Buy 800 Amps Items Including:
Extech EX730: AC/DC TRMS Clamp Meter, 800 Amps
Extech EX720: True RMS AC Clamp Meter, 800 Amps
Sight Flow Indicator, 0.5 To 6.5 GPM; PSF Body With 1/2" NPT(F) Connec (SFI-800-1/2-LF)
PTFE tubing, 8.00 x 9 mm, 25 ft/pk (YO-06363-20).
More Details
Right to Know Labels
Buy Right to Know Labels Items Including:
Right-To-Know Compliance Center, Single binder, English (2010)
Right-To-Know Compliance Center, Double binders, English (2010DB)
Right-To-Know Compliance Center, Double binders, English/Spanish (2010DBSP)
Right-to-Know training poster, 32'' X 20''.
More Details
Small Enclosures
Buy Small Enclosures Items Including:
Cole-Parmer Balance Draft Shield, Small 13"W x 13-1/2"H x 14-3/4"D (small)
Bud Industries TR-Series Enclosure for DIN-Mount Controllers, Small Si
Small 1/16 DIN-Mount Meter/Controller Enclosures (YO-94503-36).
More Details
70618
results
1
2
3
4
5
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Labpette Liquid Handling to Small Enclosures
>
31125329-s-toc_2 31125329stoc2