Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 1000

Hot Water Flow Meter   to   Chemistry Laboratory Glassware

Image
  Product Detail
More Detail

Hot Water Flow Meter

Buy Hot Water Flow Meter Items Including:
  • Cole-Parmer Flow Cell for Ultrapure Water, 316 SS
  • DO Membrane Kit, 2.00 mil PE for low flow, profiling and flow cell app (5909)
  • Flow cell for Horiba U-50 meters (3200156570)
  • YSI Model 3255 Conductivity Cell, Flow-Through Style, Glass, K=0.1 (3255).

Pyrex Vacuum Desiccator Accessories

Buy Pyrex Vacuum Desiccator Accessories Items Including:
  • Pyrex Vacuum Desiccator, 5.8L
  • Pyrex Vacuum Desiccator, 2.4L
  • Pyrex Vacuum Desiccator, 10.5L
  • Pyrex Desiccator, 10.5L
  • Pyrex Desiccator, 2.4L

Qualatrile Blue 5 Nitrile Examination Gloves

Buy Qualatrile Blue 5 Nitrile Examination Gloves Items Including:
  • Ansell TNT Blue Nitrile Gloves, Powder Free, Small
  • Ansell TNT Blue Nitrile Gloves, Powder Free, Medium
  • Ansell TNT Blue Nitrile Gloves, Powder Free, Large.

Justrite Flammable Liquid Storage Cabinet

Buy Justrite Flammable Liquid Storage Cabinet Items Including:
  • Justrite HDPE Benchtop Acid Storage Cabinet, 8 L
  • Justrite HDPE Full-size Acid Storage Cabinet, 83 L
  • Justrite Flammable Storage Safety Cabinet, 45 gallons, self-closing do
  • Justrite Flammable Storage Safety Cabinet, 60 gallons, self-closing do.

Quality Instant Ph Tester

Buy Quality Instant Ph Tester Items Including:
  • Oakton Waterproof EcoTestr™ pH 2 Pocket pH Tester
  • Oakton Waterproof pHTestr 10 Pocket pH Tester and calibration kit
  • Oakton Waterproof pHTestr 20 Pocket pH Tester and calibration kit
  • Oakton Waterproof pHTestr 30 Pocket pH Tester and calibration kit.

Vivitar Digital 10x25 Camera Binocular

Buy Vivitar Digital 10x25 Camera Binocular Items Including:
  • Digital camera binocular compound microscope, 4x/10x/40x/100x super co (DB86.515.F01 115V/60HZ)
  • Professional Binocular Microscope with built-in 2.0 Mega Pixel Digital (DC5-163)
  • Stereozoom Microscope, digital camera, 115 VAC (DX47.056.F01 115V/60HZ).

Weights of Cylindrical Steel Tubing

Buy Weights of Cylindrical Steel Tubing Items Including:
  • Pump, Sealless Drum Pump, 15 GPM or 27.8 TDH, Stainless Steel Tube, 11
  • Pump, Sealless Drum Pump, 15 GPM or 27.8 TDH, Stainless Steel Tube, 23.

8 Inch Diameter Silicone Hose

Buy 8 Inch Diameter Silicone Hose Items Including:
  • Platinum-Cured SS Wire-Reinforced Silicone Hose 0.5 Inch ID
  • Platinum-Cured SS Wire-Reinforced Silicone Hose 0.75 Inch ID
  • Platinum-Cured SS Wire-Reinforced Silicone Hose 1.0 Inch ID
  • Platinum-Cured SS Wire-Reinforced Silicone Hose 1.5 Inch ID.

3.5 Ep Ss Tubing

Buy 3.5 Ep Ss Tubing Items Including:
  • Masterflex Standard pump head for L/S 35 tubing, PC housing/SS rotor.
  • Masterflex I/P Standard pump head for I/P 82 tubing, PC housing/SS rot
  • Masterflex I/P Standard pump head for I/P 82 tubing, PPS housing/SS ro.

Flow Meter Hydrogen Gas

Buy Flow Meter Hydrogen Gas Items Including:
  • Flow meter mounting kit, shielded cable with two 9 pinsD-connectors fo
  • Flow meter mounting kit, 4 wire cable between GFM RJ11 andTIO 9 pin D-
  • Variable-speed gas piston pump, standard, 115 VAC
  • Variable-speed liquid piston pump, standard, 115 VAC.

Small Stainless Steel Funnels

Buy Small Stainless Steel Funnels Items Including:
  • Stainless steel funnel, 2 3/8 oz (YO-07263-00)
  • Stainless steel funnel, 13 oz (YO-07265-00)
  • Stainless steel funnel, 22 1/4 oz (YO-07266-00)
  • Small Lab Frame, Stainless steel
  • Justrite Safety Drum Funnel, manual closing, 1" stainless steel flame.

Professional Infrared Thermometer

Buy Professional Infrared Thermometer Items Including:
  • Fluke 63 Infrared Thermometer, Basic model (FLUKE-63)
  • Fluke 561 Infrared Thermometer, HVAC Pro model (FLUKE-561)
  • Fluke 566 Infrared Thermometer, Advanced model (FLUKE-566)
  • Fluke 568 Infrared Thermometer, Advanced High-Temperature Model (FLUKE-568).

Flasks 2000ml Pyrex 4980

Buy Flasks 2000ml Pyrex 4980 Items Including:
  • PyrexVerified Volumetric Flask 2000mL 1/pk
  • PYREX Brand 4980 flask; 125 mL, pack of 12
  • PYREX Brand 4980 flask; 250 mL, pack of 12
  • PYREX Brand 4980 flask; 300 mL, pack of 12
  • PYREX Brand 4980 flask; 1000 mL, pack of 6.

Ion Specific Electrode Fe

Buy Ion Specific Electrode Fe Items Including:
  • ISE electrode, ammonium, for controller 56105-00 (YO-27077-01)
  • ISE electrode, chloride, for controller 56105-00 (YO-27077-04)
  • ISE electrode, fluoride, for controller 56105-00 (YO-27077-06)
  • ISE electrode, lead, for controller 56105-00 (YO-27077-08).

Ambi Dex Nitrile Gloves

Buy Ambi Dex Nitrile Gloves Items Including:
  • Best Nitri-Dex Nitrile Glove, Size 10, Package of 25 pair
  • Best N-DEX Nitrile Exam Gloves, Powder-Free, medium, 100/pack
  • Best N-DEX Nitrile Exam Gloves, Powder-Free, large, 100/pack
  • Best N-DEX Nitrile Exam Gloves, Powder-Free, extra-large, 100/pack.

Industrial Liquid Filtration Systems

Buy Industrial Liquid Filtration Systems Items Including:
  • Multiprobe Flowblock Sampling System (YO-35434-90)
  • Sludge Judge II Sampler System
  • "Big Blue" industrial filter housing; 10"L, 1" NPT(F) (150238)
  • "Big Blue" industrial filter housing; 10"L, 1" NPT(F) (150237).

Inline Stainless Butterfly Valve

Buy Inline Stainless Butterfly Valve Items Including:
  • Stainless steel liquid flow switch for high inline pressures; 0.25 GPM (YO-32774-40)
  • Stainless steel liquid flow switch for high inline pressures; 0.5 GPM (YO-32774-42).

Bowl Lift Design Stand Mixers

Buy Bowl Lift Design Stand Mixers Items Including:
  • Industrial Mixer Lift Stand, Manual (YO-04600-10)
  • Industrial Mixer Lift Stand, Winch (YO-04600-20).

Napa Barbed Brass Fitting Adapter

Buy Napa Barbed Brass Fitting Adapter Items Including:
  • Luer fittings, Nickel-plated brass fitting, male slip x 1/4" barb (YO-31507-60)
  • Nickel-plated brass fittings, male luer lock x 1/16" hose barb (YO-31507-62)
  • Barbed male threaded adapter, PVDF, 3/4 x 5/8" NPTM, 25/pack (YO-30625-09).

Personal Protective Equipment Program

Buy Personal Protective Equipment Program Items Including:
  • Safety Sign, Caution - Personal Protective Equipment Required Beyond T (MPPE796VS)
  • Safety Sign, Caution - Personal Protective Equipment Required Beyond T (MPPE796VP)
  • Safety Sign, Caution - Personal Protective Equipment Required Beyond T (MPPE796VA).

Dual Adjustable Dc Power Supplies

Buy Dual Adjustable Dc Power Supplies Items Including:
  • Instek Model SPD-3606 Multiple Output, Dual Range, DC Power Supply, 37 (SPD-3606)
  • B&K Precision 9171 Programmable Dual Range DC Power Supply, 10V/10A, 2
  • B&K Precision 9172 Programmable Dual Range DC Power Supply, 35V/3A, 70.

Potato Dextrose Agar Fungi

Buy Potato Dextrose Agar Fungi Items Including:
  • Cole-Parmer Potato dextrose agar, 500 g jar
  • Prepared agar plate, PDA (potato dextrose agar), 10/pk
  • Potato Dextrose Agar 500g
  • Thermo Scientific Potato dextrose agar, pH 5.6, 10/pk.

Labtop Rotary Lobe Pump

Buy Labtop Rotary Lobe Pump Items Including:
  • Sanitary Wash-Down Rotary Lobe Pump, 13 GPM, 230 VAC, Single-phase
  • Sanitary Wash-down Rotary Lobe Pump, 26 GPM, 115 VAC, Single-phase
  • Sanitary Wash-down Rotary Lobe Pump, 36 Gpm, 115 Vac, Single-phase (no.

Qorpak Kaptclean Glass Sample Jars

Buy Qorpak Kaptclean Glass Sample Jars Items Including:
  • Qorpak® Wide-mouth Sample Jar, 480 mL, Pk Of 4 (GLC05257)
  • Qorpak Wide-mouth Sample Jar, 120 mL, Cs Of 24
  • Qorpak Wide-mouth Sample Jar, 240 mL, Cs Of 24
  • Qorpak Wide-mouth Sample Jar, 480 mL, Cs Of 12
  • Qorpak Wide-mouth Sample Jar, 960 mL, Cs Of 12.

Gas Line Flexible Tubing

Buy Gas Line Flexible Tubing Items Including:
  • Tygon FEP-Lined Tubing, Non-DEHP, 1/8"ID x 1/4"OD, 50 ft/pack
  • Tygon FEP-Lined Tubing, Non-DEHP, 3/16"ID x 5/16"OD, 50 ft/pack
  • Tygon FEP-Lined Tubing, Non-DEHP, 3/8"ID x 9/16"OD, 50 ft/pack.

White Heat Shrink Tubing

Buy White Heat Shrink Tubing Items Including:
  • Heat-shrink tubing assortment pack, polyolefin
  • Heat-shrink tubing assortment pack, PVC
  • Double-walled PTFE heat-shrink tubing, 0.065"ID, 12 ft/pack (YO-95810-10)
  • Double-walled PTFE heat-shrink tubing, 0.115"ID, 12 ft/pack (YO-95810-11).

Activated Carbon Air Capsule Filter

Buy Activated Carbon Air Capsule Filter Items Including:
  • Double open end granular activated carbon cartridge; 20 µm, 10"L (155155-43)
  • Double open end granular activated carbon cartridge; 20 µm, 20"L (155138-43)
  • Parker Domnick Hunter Compressed Air Activated Carbon Filter Element, (YO-29587-21).

Adjustable Stem Float Switch

Buy Adjustable Stem Float Switch Items Including:
  • LS-1700 Series Level Switch, 316ss Stem Buna-N Float 20VA (01702).

High Pressure Pump 6 4

Buy High Pressure Pump 6 4 Items Including:
  • Masterflex L/S High-Pressure Pump System; 1 to 100 rpm, 90 to 260 VAC
  • Pump, High Precision - Ultra High Pressure Pumps, up to 18,000 psi, Co (F05SNXP1-SP1)
  • Ismatec™ high-pressure pump with Series Q pump heads, 0.016 to 19 m (MAKE-UP) (YO-78021-00).

Chemistry Laboratory Glassware

Buy Chemistry Laboratory Glassware Items Including:
  • General Lab Glassware Starter Kit with Tote.



Hot Water Flow Meter to Chemistry Laboratory Glassware

>