Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 11
Platform Dollies
to
Ionplus Ion Selective Electrodes
Image
Product Detail
More Detail
Platform Dollies
Buy Platform Dollies Items Including:
Enpac Cylinder dolly
Rubbermaid Drum Dolly For 55 Gal Drums
Brute Container Dolly.
More Details
Hand Water Pumps
Buy Hand Water Pumps Items Including:
Water Pump, 6 strokes/gallon, 3 foot inlet tube
Pump, Motorized Drum Pump Package, Designed for Water Treatement Chemi (9430-110V) (YO-70614-00)
Pump, Motorized Drum Pump Package, Designed for Water Treatement Chemi (9431-230V) (YO-70614-01).
More Details
Compact Dew
Buy Compact Dew Items Including:
Temperature & Humidity and Dew Point Data Logger with LCD Display, 16,
Increased accuracy tmperature, humidity, and dew point USB Data Logger.
More Details
Extech Digital Sound Level Meter
Buy Extech Digital Sound Level Meter Items Including:
Extech Digital Sound Level Meter
Extech Type 2 Digital Sound Level Meter
Extech Type 2 Digital Sound Level Meter Kit
Extech Digital Sound Meter with NIST Cert
Extech 407768 Digital Sound Meter.
More Details
Amprobe Light Meters
Buy Amprobe Light Meters Items Including:
Amprobe LM-100 Digital Light Meter
Amprobe LM-120 Autoranging Digital Light Meter.
More Details
Bottles Detergent
Buy Bottles Detergent Items Including:
Citranox Cleaning Detergent, case of four 1-gallon bottles
Luminox Cleaning Detergent, case of four 1-gallon bottles
Liqui-Nox Cleaning Detergent, case of four 1-gallon bottles
Liqui-Nox cleaner for general purpose cleaning, 1 quart bottle, case o.
More Details
Digital Stereo
Buy Digital Stereo Items Including:
Stereozoom Microscope, built-in 2.0 mega pixel digital camera, 110-220 (DX53056101).
More Details
Tygothane Tubing
Buy Tygothane Tubing Items Including:
Tygothane pressure tubing, 3/4"ID x 1-1/16"OD, 100 ft/roll
Tygothane pressure tubing, 1-1/4"ID x 1-3/4"OD, 50 ft/roll
Pumps, Metering Pumps Tygothane Replacement Tube Set for High-Flow Ran (YO-74204-56).
More Details
Modular Pumps
Buy Modular Pumps Items Including:
Masterflex L/S Precision Modular Pump System with Easy-Load 3 Pump Hea
Masterflex L/S Precision Modular Pump System with Easy-Load II Pump He
Masterflex I/P Precision Modular Pump System with Easy-Load Pump Head;.
More Details
Lab on a Chip Technology
Buy Lab on a Chip Technology Items Including:
Microfluidic Fluid Splitter Slide, Several splitters on one slide
Microfluidic Snake Mixer Slide, COC material, several mixers on one ch
Microfluidic Microplat Assembly Platform
Microfluidic Micro Luer Connector, COC material, package of 25 pieces.
More Details
Card Dual
Buy Card Dual Items Including:
Keithley Instruments 3720 Multiplexer card, dual, 1 x 30 channels, aut
Keithley Instruments 3721 Multiplexer card, dual, 1 x 20 channels, aut
Keithley Instruments 3722 Multiplexer card, dual, high density, 1 x 48.
More Details
Autoradiography Film
Buy Autoradiography Film Items Including:
Cole-Parmer Blue-Sensitive Autoradiography Film, 8 x 10", 100/pk
Autoradiography Cassette; aluminum, 8" x 10" (YO-21700-34)
Autoradiography Cassette; stainless steel, 5" x 7" (YO-21700-40)
X-ray Film Blue 14x17" 100pk.
More Details
Brady Lockout Tagout
Buy Brady Lockout Tagout Items Including:
Brady Personal Valve and Electrical Lockout Kit
Brady Ready Access Padlock Station
Brady Personal Lockout Kit in Belt Pouch
Brady Personal Electrical Lockout Kit in Plastic Case.
More Details
Adhesive Mats
Buy Adhesive Mats Items Including:
Adhesive Mat Starter Kit White 30 Layer
Adhesive Mat Starter Kit Blue 30 Layer
Adhesive Mat Starter Kit Blue 60 Layer
Mat Refill 30 Layer Blue Pk 4
Mat Refill 60 Layer Blue Pk 4
More Details
Honeywell Process Equipment & Instrumentation
Buy Honeywell Process Equipment & Instrumentation Items Including:
Honeywell UDC 700 Universal-Input Controller, 90-264V.
More Details
Fluke Voltage
Buy Fluke Voltage Items Including:
Fluke Voltage Detector, 90-1000 volts AC
Fluke Voltage Detector, 24-90 volts AC
Fluke 62/322/1AC IR Thermometer, Clamp Meter, and Voltage Detector Kit (FLUKE-62/322/1AC)
Fluke-179/1AC-II Multimeter, Voltage Tester & Acc
Kit (FLUKE-179/1AC-II).
More Details
Carts Utility
Buy Carts Utility Items Including:
Stainless Steel Equipment Cart without drawers; 18-3/8" x 16-5/8", 3 s
SS equipment cart without drawers; 18-3/8" x 16-5/8", 3 shelves, 400 l
SS equipment cart with drawers; 18-3/8" x 16-5/8", 3 shelves, 200 lb c (356).
More Details
Glasses Brand
Buy Glasses Brand Items Including:
Cole-Parmer Glass Body Fermentation DO Probe, 180mm immersion depth, p
Pyrex Brand 3024 cylinder; 5 mL, pack of 1
Pyrex Brand 3024 cylinder; 10 mL, pack of 1
Pyrex Brand 3024 cylinder; 25 mL, pack of 1
Pyrex Brand 3024 cylinder; 100 mL, pack of 1.
More Details
Cole-Parmer Dye 26
Buy Cole-Parmer Dye 26 Items Including:
Solvent-based Flouresent RED Dye, 16 OZ.
More Details
Salt in Crude
Buy Salt in Crude Items Including:
Oakton EcoTestr Salt pocket salinity tester
Oakton SALT 6+ handheld salinity meter kit
Oakton SALT 6+ handheld salinity meter with probe and NIST-traceable C
Low-cost salt refractometer, 0 to 28% salinity
Atago Salt/specific gravity refractometer.
More Details
2 Bar Pump
Buy 2 Bar Pump Items Including:
Etatron DLX pH/ORP Pump Control System, 2 L/hr, 10 bar max pressure, w (DLX-pH-RX/MBB) (YO-56022-41)
Etatron DLXB pH/ORP Pump Control System, 2 L/hr, 10 bar max pressure, (DLXB-PH-RX/MBB) (YO-56022-48).
More Details
Dpi 620
Buy Dpi 620 Items Including:
GE (Druck) DPI 620 Multifuction Calibrator (Now capable of HART Commun
IDOS to DPI 620 Adaptor
DPI 620 Intecal field calibration manager (IO620-FIELD-CAL)
DPI 620 system carry case (IO620-CASE-2)
DPI 620 Modular system transit case (IO620-CASE-4).
More Details
20 Dual
Buy 20 Dual Items Including:
Cole-Parmer Vertical Dual Adjustable Slab Gel System
Cole-Parmer Vertical Dual Adjustable Slab Gel System, CE-marked
ThermoProbe TL2 Dual Channel RTD Benchtop Thermometer (Two Probes)
3M BX™ Dual Reader Safety Glasses, 2.0X top and bottom diopters.
More Details
Ptfe Vessel
Buy Ptfe Vessel Items Including:
Glass Vessel; 2 mL (011.102)
Glass Vessel; 5 mL (011.105)
Glass Vessel; 10 mL (011.110)
Glass Vessel; 15 mL (011.115)
Glass Vessel; 30 mL (011.130).
More Details
Balances Mettler Toledo
Buy Balances Mettler Toledo Items Including:
Mettler Toledo Excellence Plus XP Micro Balance 5.1/22G X .001MG/.01MG
Mettler Toledo Excellence Plus XP Semi-Micro Balance, 31/120g X 0.01/0
Mettler Toledo Excellence Plus XP Precision Toploading Balance, 1210g (XP1203S).
More Details
Pack Label
Buy Pack Label Items Including:
Address Label, 130 Labels Per Roll, Two Roll/Pack
Shipping Label, 300 Labels Per Roll
One Roll/Pack
Name Badge, 250 Labels Per Roll, One Roll/Pack
Color Change Crayon Kit, 10/pack (YO-90307-50)
More Details
Computer Connecting Cable
Buy Computer Connecting Cable Items Including:
Connecting cable ZEN V2 to computer/laptop, 2 meter (see attached spec
RS-232 interface cable; connects L/S computer-compatible drives to PC, (YO-07550-64).
More Details
Ohaus Mb45
Buy Ohaus Mb45 Items Including:
Ohaus In-Use Cover for MB45 Balances
Ohaus MB Moisture Determination Balance, Mb45 45 gX 0.001 G 120VAC.
More Details
Mist Bottles
Buy Mist Bottles Items Including:
Wheaton Chromatography Fine-Mist Spray Bottle (YO-34695-22)
Quick Mist Dispenser 16 Oz.
More Details
Ionplus Ion Selective Electrodes
Buy Ionplus Ion Selective Electrodes Items Including:
Thermo Scientific Orion bromide electrode ionplus Sure-Flow with WP BN
Thermo Scientific Orion cupric electrode ionplus Sure-Flow with WP BNC
Thermo Scientific Orion cyanide electrode ionplus Sure-Flow with WP BN.
More Details
70618
results
<<<Previous
7
8
9
10
11
12
13
14
15
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Platform Dollies to Ionplus Ion Selective Electrodes
>
31125329-s-toc_2 31125329stoc2