Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 13
2000 Ml Flask
to
Pyrex Plates
Image
Product Detail
More Detail
2000 Ml Flask
Buy 2000 Ml Flask Items Including:
Erlenmeyer flask mantle for 2000-ml flask, 400 watts, 230 VAC (100AO929)
Spherical heating mantle for 2000-ml flask, 500 watts lower half, 200 (100A O1103)
Fast-Freeze flask, 2000 mL
Thermo Scientific Nalgene polypropylene filtering flask, 2000 mL.
More Details
Freeze Dryer System
Buy Freeze Dryer System Items Including:
12L Console Freeze Dryer With Purge Valve And Mini Chamber 230vac 60hz (YO-03336-73)
12L Console Freeze Dryer With Purge Valve And Mini Chamber 220vac 50hz (YO-03336-63)
1 L Benchtop Freeze Dry System, -54° with PTFE interior, 115V (YO-03337-05).
More Details
Reagent Grade Water
Buy Reagent Grade Water Items Including:
Water (Environmental Grade) (1 liter) (YO-88042-46)
Water (Biotech Grade) (20 liter) (YO-88059-63)
Water, ECD tested for pesticide analysis (2.5l) (YO-88294-46)
Water, DNase, RNase, Protease free (1l) (YO-88295-49).
More Details
Range Hoods Stainless
Buy Range Hoods Stainless Items Including:
12x10x6 Type 304 Junction Box, Stainless Steel (JB121006H4)
12x10x6 Type 316L Junction Box, Stainless Steel (JB121006H6)
12x12x6 Type 316L Junction Box, Stainless Steel (JB121206H6)
14x12x6 Type 316L Junction Box, Stainless Steel (JB141206H6).
More Details
Raytek Infrared Thermometers
Buy Raytek Infrared Thermometers Items Including:
Raytek infrared thermometer (RAYMT4U)
Comapact CM-Series Infrared Sensor Type-J T/C Output with 3' Cable (RAYCMLTJ)
Mi3 Sensor, Standard Model, 10:1, -40-600°C, 3m Cable
Sensor head mounting nuts (5 pieces)
Sensor head adjustable mounting bracket.
More Details
Lamotte Test Kits
Buy Lamotte Test Kits Items Including:
Soil texture test kit (1067)
Garden test kit (5679)
Soil Nutrient Test Kit; Macronutrient and pH (5029)
LaMotte Dissolved Oxygen Test Kit, 50 tests/kit
Aluminum test kit (3641-SC)
More Details
Boxes and Storage
Buy Boxes and Storage Items Including:
Polystyrene Trans-Boxes container; 6 sections, 1-5/8" x 6" x 1-1/2", 2
Polystyrene Trans-Boxes container; 18 sections, 1-5/8" x 1-7/8" x 1-1/
K-resin Trans-Boxes container; 24 sections, 2" x 2" x 2", 12/pack.
More Details
Large Tube
Buy Large Tube Items Including:
Thermo Scientific Labquake Tube Shaker/Rotators, Large, 32 Tubes, 10 m (415110Q)
Barbed fitting kits, Barbed connector/reducer kits, Natural PP, large (YO-31514-07)
Barbed elbow/tee/Y-connector large fittings kits; Natural PP material (YO-31514-19).
More Details
Labnet Shakers
Buy Labnet Shakers Items Including:
Microplate Orbital Shaker with Timer, 230 VAC, 50 Hz
Additional Tube Rack/Workstation for 17310-50 and -52 (YO-17310-54)
Adapter F 0.5/0.6ml Tubes - 6/pk (YO-17310-58)
Microplate/Microtube Shaking Incubator - 120VAC (YO-17310-50).
More Details
Adam Module
Buy Adam Module Items Including:
12 Channel Sourcing Iso Digital Output Data Aquisition Module (Modbus)
8 Channel Relay Output Module (Modbus) (ADAM-4068-BE)
Isolated RS-232 to RS-422/485 Converter (ADAM-4520-D2E)
2-port Ethernet to RS-232/485/422 ComPort Server (ADAM-4570-BE).
More Details
Poly Tubing
Buy Poly Tubing Items Including:
Poly-Cone™ tapered tubing adapter (YO-06432-00)
Male luer integral lock ring to 1/8" semi-rigid tubing hose barb, poly (YO-45503-11).
More Details
Data Acquisition Equipment
Buy Data Acquisition Equipment Items Including:
Keithley Instruments 2700 Multimeter, Data Acquisition System with 2 S
Keithley Instruments 2701 Multimeter, Data Acquisition System with 2 S
Keithley Instruments 2750 Multimeter, Data Acquisition Switch System w.
More Details
Temperature Plates
Buy Temperature Plates Items Including:
RET basic IKAMAG Stirring Hot Plate 115V (3622001)
ENVIRO-SAFE POCKET THERMOMETER, NICKEL PLATED BRASS CASE, -10/110C, 16 (20568B)
EASY-READ POCKET THERMOMETER, WINDOW NICKLE-PLATED BRASS CASE, -5/50C
ETS-D5 Temperature Probe (YO-04671-55).
More Details
Cole-Parmer Digital Specific Gravity Meters
Buy Cole-Parmer Digital Specific Gravity Meters Items Including:
Low-Cost Specific Gravity Meter, 0.750 to 1.000 SGU, °C model (DSG-11-075100-C-B)
Low-Cost Specific Gravity Meter, 1.000 to 1.300 SGU, °C model (DSG-11-100130-C-B)
Digital Density / Specific Gravity Meter
SBS-2002 : Digital Hydrometer (SBS-2002).
More Details
Half Mask Respirator
Buy Half Mask Respirator Items Including:
North Half-Mask Respirator, Medium
Survivair Valuair Plus half-mask respirators, Medium
Survivair® Valuair® Plus half-mask respirators, Large (YO-81661-05)
Survivar Premier Half-Mask Respirator, Medium
Survivar Premier Half-Mask Respirator, Large.
More Details
Parker Hannifin Male Pipe Adapters (Straight)
Buy Parker Hannifin Male Pipe Adapters (Straight) Items Including:
Compression fittings, male pipe adapter, straight, Black PP, 1/4" OD x (YO-06444-01)
Compression fittings, male pipe adapter, straight, Black PP, 3/8" OD x (YO-06444-02).
More Details
Digital Multimeter Tester
Buy Digital Multimeter Tester Items Including:
Digital Multi Tester, DM78C (DM78C)
Dranetz Insulation Tester/TRMS Digital Multimeter/Datalogger
Fluke-179/1AC-II Multimeter, Voltage Tester & Acc
Kit (FLUKE-179/1AC-II)
Fluke-62 MAX+/T+PRO/1AC Electrical/HVAC Combo Kit.
More Details
Oakton Multiparameter Pocket Testers
Buy Oakton Multiparameter Pocket Testers Items Including:
Oakton PTTestr 35 Waterproof Multiparameter pH/TDS pocket tester
Oakton PCTestr 35 Waterproof Multiparameter pH/conductivity pocket tes.
More Details
Cole-Parmer Silver Chloride
Buy Cole-Parmer Silver Chloride Items Including:
Silver Electrodes, Two Detecting Electrodes, One Cathode (YO-02656-32).
More Details
Labconco Glassware Carts with Wire Basket
Buy Labconco Glassware Carts with Wire Basket Items Including:
Glassware cart with four small wire baskets (8045000).
More Details
Oakton Fep-Insulated-Wire Probes
Buy Oakton Fep-Insulated-Wire Probes Items Including:
Type J FEP-insulated thermocouple probe with PTFE-coated junction (072097B-J)
Type K FEP-insulated thermocouple probe with PTFE-coated junction (072097B-K)
Type T FEP-insulated thermocouple probe with PTFE-coated junction (072097B).
More Details
Dc Speed Controller
Buy Dc Speed Controller Items Including:
Basic Variable-Speed DC Motor Controller for 1/4 to 2 hp motors.
More Details
High Temperature Bath
Buy High Temperature Bath Items Including:
High-Stability Immersion Digital Circulator; 120V; 50/60Hz (YO-01262-05).
More Details
Material Handling Hand Trucks
Buy Material Handling Hand Trucks Items Including:
Rubbermaid Platform Truck, 36" x 24", 500 lb capacity.
More Details
Tubing Peristaltic
Buy Tubing Peristaltic Items Including:
Tube Assembly Kit for the peristaltic pump 74130-60 and 74130-61 pumps
Pumps, Metering Pumps Replacement Tube Set for Mid-Flow Range Cole-Par (YO-74204-01)
Pumps, Metering Pumps Replacement Tube Set for Mid-Flow Range Cole-Par (YO-74204-02).
More Details
Handheld Viscometer
Buy Handheld Viscometer Items Including:
Handheld Mini 254 nm Ultraviolet Lamp (95-0158-04(UVG-4))
Handheld Mini 254/365 Switchable UV Lamp (95-0188-02(UVSL-14P))
Viscometer Tube, Stainless steel, for Saybolt Viscometer Bath (K22309)
Displacement Ring for Saybolt Viscometer Bath (K22080).
More Details
Cole-Parmer Polycarbonate Luer Fittings and Adapters
Buy Cole-Parmer Polycarbonate Luer Fittings and Adapters Items Including:
Luer fittings, Polycarbonate fittings, male lock x male lock, rotating (YO-06464-90)
Luer fittings, Polycarbonate fittings, female x male lock, rotating, (YO-06464-95)
Cole-Parmer Animal Free Female Luer Fittings, 1/8", PP, 25/pk (YO-30800-02).
More Details
Track Temperature
Buy Track Temperature Items Including:
Track It™ temperature data logger with display and standard battery
Track It™ temperature and humidity data logger with display and sta (YO-30003-02)
Track It™ temperature and humidity data logger with display and lon (YO-30003-03).
More Details
Danger Tag
Buy Danger Tag Items Including:
Tag, Danger Do Not Operate This Tag And Lock(bilingual Spanish), 5 7/8 (TSP101CTP)
Tag, Danger Do Not Operate This Tag And Lock (bilingual Spanish), 5 7/ (TSP101PTP)
Tag, Danger Do Not Operate This Tag And Lock (bilingual Spanish), 5 3/ (TSP101LTP).
More Details
Pyrex Plates
Buy Pyrex Plates Items Including:
Pyrex Brand 9820 culture tube; 11 mL, pack of 72
Pyrex Brand 9820 culture tube; 15 mL, pack of 72
Pyrex Brand 9820 culture tube; 20 mL, pack of 72
Pyrex Brand 9820 culture tube; 27 mL, pack of 72
Pyrex Brand 9820 culture tube; 34 mL, pack of 72.
More Details
70618
results
<<<Previous
9
10
11
12
13
14
15
16
17
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,354
2000 Ml Flask to Pyrex Plates
>
31125329-s-toc_2 31125329stoc2