Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 15
Top Safety Products
to
Seal Plate
Image
Product Detail
More Detail
Top Safety Products
Buy Top Safety Products Items Including:
Counter top mounted combination eyewash/drench hose with stay-open val (G5022)
Wearwell Tile-Top Antifatigue Mats, 24" x 36", 1/2"H, black and white
Wearwell Ultrasoft Tile-Top Antifatigue Mats, 24" x 36", 7/8"H, black.
More Details
Precision Hot Plate
Buy Precision Hot Plate Items Including:
Cole-Parmer Five-Position Stirring Hot Plate, 120 VAC
EchoTherm™ Digital Hot Plate, Ceramic, 115 VAC
EchoTherm™ Digital Hot Plate, Aluminum, 115 VAC
EchoTherm™ Digital Hot Plate, Aluminum, 230 VAC
EchoTherm™ Digital Stirring Hot Plate , 115 VAC.
More Details
Wika Transmitter
Buy Wika Transmitter Items Including:
F-20-3A : Transmitter, 100psi, 4-20MA Sanitary 3A Pressure Transmitter (YO-68866-08)
F-20-3A : Transmitter, 200psi, 4-20MA Sanitary 3A Pressure Transmitter (YO-68866-10)
Wika Pressure Transmitter ~ 0 To 5 Psi, 1/2" NPT(m); 4 To 20 mA Output.
More Details
Contact Relays
Buy Contact Relays Items Including:
Module 2 External Dry Contact Relays
Contact Relay Module.
More Details
A&D Weighing Viscometers
Buy A&D Weighing Viscometers Items Including:
Low range tuning fork vibration viscometer 120 VAC
High range tuning fork vibration viscometer 120 VAC
High range tuning fork vibration viscometer 240 VAC.
More Details
Kimcote Filtering
Buy Kimcote Filtering Items Including:
Kimcote, Heavy-Wall, Safety-Coated, Filter Flask, 125mL (YO-34524-49)
Kimcote, Heavy-Wall, Filter Flask, 1000mL (YO-34524-51)
Kimcote, Heavy-Wall, Filter Flask, 2000mL (YO-34524-53).
More Details
Microbial Contamination
Buy Microbial Contamination Items Including:
Sterile two-piece monitor, gridded surface, 0.45 um, 50/pack (YO-02935-20)
Mixed Cellulose Ester (MCE) membrane for asbestos monitoring; 25-mm di (YO-36230-30)
Mixed Cellulose Ester (MCE) membrane for asbestos monitoring; 37-mm di (YO-06639-72).
More Details
Ge Sensing Modules
Buy Ge Sensing Modules Items Including:
PM 620 Pressure Module +/-10 inH2O psi, gauge type (PM 620 008G)
PM 620 Pressure Module +/-1 psi, gauge type (PM 620 01G)
PM 620 Pressure Module +/-10 psi, gauge type (PM 620 04G)
PM 620 Pressure Module -14.5 to 100 psi, gauge type (PM 620 10G).
More Details
Cole-Parmer Tellurium
Buy Cole-Parmer Tellurium Items Including:
TELLURIUM POWDER -200M 100G (YO-88288-70).
More Details
Nalgene Water and Industrial Liquid Sampling
Buy Nalgene Water and Industrial Liquid Sampling Items Including:
Thermo Scientific Nalgene Storm Water Sampler Bottle, HDPE for Inorgan
Thermo Scientific Nalgene Storm Water Sampler Bottle, Amber Glass for.
More Details
Cga 580 Fitting
Buy Cga 580 Fitting Items Including:
Single-Stage Regulator, 2000 scfh Capacity, 580 CGA Fitting (YO-98202-03)
Single-Stage Regulator, 3500 scfh Capacity, 580 CGA Fitting (YO-98202-13)
Single-stage Regulator, 270 scfh Capacity, 580 CGA fitting (YO-98210-26).
More Details
Cole-Parmer Chamber Accessories
Buy Cole-Parmer Chamber Accessories Items Including:
HMK 15A1010A01B : RH Cal Kit with 2 Standard Chambers LICL and NACL (YO-03335-10)
Clear Drying Chambers (YO-03337-64)
8 Port Clear Drying Chambers (YO-03337-66)
Drying chamber with 12 ports (YO-03346-01)
Drying chamber with 16 ports (YO-03346-11).
More Details
Probe Remote
Buy Probe Remote Items Including:
Remote Monitoring RTD Thermometer
Sound Meter With Remote Probe
Deluxe Remote Probe Thermometer (4377CP)
Waterproof Remote Probe Thermometer (4039CP)
Stick Style Remote Probe Thermometer °C (4144CP)
More Details
Filter Felt
Buy Filter Felt Items Including:
Size 1 high-flow polyester filter bag; 300°F (149°C) maximum (255046-03)
Size 1 high-flow polyester filter bag; 300°F (149°C) maximum (255047-03)
Size 3 high-flow polypropylene filter bag; 200°F (93°C) maximu (255078-03).
More Details
Tools Micrometer
Buy Tools Micrometer Items Including:
52-229-201:1in Economy Micrometer (52-229-201)
Battery-powered digital micrometers with LCD and RS-232 output, 0 to 1 (54-850-001)
Micrometer with digital counter, 0 to 1" range (52-222-001-1)
54-860-777:Micrometer Inspection Set (54-860-777).
More Details
Plasteel Furniture & Fume Hoods
Buy Plasteel Furniture & Fume Hoods Items Including:
Starter four-shelf vented units, 36"L x 18"W
Starter four-shelf vented units, 48"L x 18"W
Starter four-shelf vented units, 60"L x 18"W
Starter four-shelf vented units, 36"L x 24"W.
More Details
Product Chiller
Buy Product Chiller Items Including:
Cole-Parmer Flow-through Refrgerated Chillers; range, -25 to 40°C; (YO-01283-40)
Cole-Parmer Flow-through Refrgerated Chillers; range, -25 to 40°C; (YO-01283-41)
Polyscience turbine low temp benchtop chiller, -20 to 40°C, 500 wa.
More Details
Nilfisk 80
Buy Nilfisk 80 Items Including:
Nilfisk Lightweight Cleanroom Vacuum, 240 VAC (01790432 (GM 80) 220V).
More Details
Venturi Pumps
Buy Venturi Pumps Items Including:
Venturi vacuum pump, 0.5 cfm, 28.0
Venturi vacuum pump, 1.2 cfm, 28.0
Venturi vacuum pump, 3.2 cfm, 28.0
Miniature venturi vacuum pump, 0.5 cfm, 28.0
VENTURI with AIR SAVER technology,28" Hg,1.2 CFM, With Gauge.
More Details
1.3 Megapixels
Buy 1.3 Megapixels Items Including:
Digital Microscope, 1.3 megapixels, 10x to 200x magnification, USB con (26700-200)
Digital Microscope, 1.3 megapixels, 500x magnification, USB connection (26700-204)
Digital Ultraviolet Microscope, 1.3 megapixels, 10x to 200x magnificat (26700-205).
More Details
Nunc 96-Well Plates
Buy Nunc 96-Well Plates Items Including:
Thermo Scientific Nunc™ 96-Well Microplates, Immunology, PS, Standa (YO-01928-22)
Thermo Scientific Nunc™ 96-Well Microplates, Immunology, PS, Standa (YO-01928-32)
Thermo Scientific Nunc 96-Well Microplates, Nontreated, PP, Conical pl (YO-01929-30).
More Details
Cole-Parmer Turntables
Buy Cole-Parmer Turntables Items Including:
1-level turntable, white
2-level turntable, white
Three-level turntable, white
Turntable, 10" Dia
Turntable, 12" Dia.
More Details
Cole-Parmer Cotton Inspection Gloves
Buy Cole-Parmer Cotton Inspection Gloves Items Including:
100% Cotton Inspection Gloves, women's, 8"L, 12 pairs/pk
100% Cotton inspection gloves, men's, 9"L, 12 pairs/pk
Inspection Glove, Cotton, Mediumweight, Mens, 12 pairs/pk.
More Details
Cole-Parmer Motorized Cap Torque Testers
Buy Cole-Parmer Motorized Cap Torque Testers Items Including:
Imada MTG-15 Digital Cap Torque Tester, 15 lbf.
Imada MTG-40 Digital Cap Torque Tester, 40.00 lbf..
More Details
Sieves 14 in Diameter
Buy Sieves 14 in Diameter Items Including:
Cole-Parmer Testing Sieve, 8" OD Brass Frame/Brass Wire, Full height, (YO-59980-03)
Cole-Parmer Sieve with NIST-Traceable Calibration Report, SS Frame/SS (YO-59984-53)
Cole-Parmer 3"-Diameter Sieve, Full Height Stainless Steel, No
14 (1..
More Details
Tissue Culture Lab
Buy Tissue Culture Lab Items Including:
Tissue Culture Filters
Pack of five
(4-000-051)
Heraeus Megafuge 40 120V Tissue Culture Bundle 40/88 (includes below p
Heraeus Megafuge 40R (Refrigerated) 120V Tissue Culture Bundle 40/88 (
More Details
Suction Equipment
Buy Suction Equipment Items Including:
Suction-feed powder-liquid dispersing mixing head for 04300-01 Magic L (R075262)
Cole-Parmer Polystat recirculator with force/suction pump, 21 lpm, 50 (YO-13042-21)
Cole-Parmer Polystat recirculator with force/suction pump, 21 lpm, 50 (YO-13042-23).
More Details
Paddle Blenders
Buy Paddle Blenders Items Including:
Paddle Blender; 50 To 400 mL, Variable Speed, Window Door, Removable P (YO-36850-13)
Paddle Blender; 200 To 3500 mL, Variable Speed, Stainless Steel Door
PADDLE BLENDER STARTER KIT; 400 ML Fixed speed blender with transparen (YO-36850-17).
More Details
Large Rack
Buy Large Rack Items Including:
Foam Test Tube Rack-large, 6 per pack
Bud Industries TR-Series Enclosure for DIN-Mount Controllers, Large Si
Base, Large V (53-2352W)
SHUR/Clean™ Reagent Bowl Rack, Large
DIN-Mount Meter/Controller Enclosures, With 1 1/16 DIN Cutout (YO-94503-54).
More Details
Seal Plate
Buy Seal Plate Items Including:
Cole-Parmer PCR Tubes and Plates Sealing Film, 100/pk
Corning cell culture flask, 25 cm2, canted neck, plug seal
Corning cell culture flask, 75 cm2, canted neck, plug seal
Corning cell culture flask, 150 cm2, canted neck, plug seal.
More Details
70618
results
<<<Previous
11
12
13
14
15
16
17
18
19
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Top Safety Products to Seal Plate
>
31125329-s-toc_2 31125329stoc2