Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 17
Hvac
to
Colder Npt(M) Threaded Adapter Inserts
Image
Product Detail
More Detail
Hvac/R Heat Sealers with Cutter
Buy Hvac/R Heat Sealers with Cutter Items Including:
Heat sealer with built-in cutter, 12", 230 VAC (AIE-305C2 (220V)).
More Details
Acs Reagent
Buy Acs Reagent Items Including:
CHROMIUM TRIOX CERT ACS 21/2KG (YO-88050-06)
CITRIC ACID GRAN CERT ACS 10KG (YO-88050-07)
CITRIC AC MONOHYD USP/FCC 10KG (YO-88050-11)
CITRIC AC MONOHYD USP/FCC 3KG (YO-88050-12)
PERCHLORIC AC 60% ACS 4X7LB/CS (YO-88051-01).
More Details
Sweep Function Generator
Buy Sweep Function Generator Items Including:
B&K Precision 4014B Sweep Function Generator, 12 MHz
BK Precision 4040B Sweep Function Generator, 20 MHz.
More Details
Fuel Hose
Buy Fuel Hose Items Including:
Tygon fuel and lubricant tubing, 3/32"ID x 3/16"OD, 50 ft/pack
Tygon fuel and lubricant tubing, 3/16"ID x 5/16"OD, 50 ft/pack
Tygon fuel and lubricant tubing, 5/16"ID x 7/16"OD, 50 ft/pack.
More Details
Thermo Scientific Jars
Buy Thermo Scientific Jars Items Including:
Thermo Scientific Nalgene polycarbonate vacuum plate for 06761-25, -30
Thermo Scientific Nalgene polypropylene wide-mouth jar, 2 L
Thermo Scientific Nalgene polypropylene wide-mouth jar, 4 L
Thermo Scientific Nalgene polypropylene jar, 2 1/4 quart.
More Details
International Cryogenics
Buy International Cryogenics Items Including:
Thermo Scientific Nunc CryoTube Starfooted Cryogenic Vials with intern (377224)
Thermo Scientific Nunc CryoTube Starfooted Cryogenic Vials with intern (377267)
Thermo Scientific Nunc CryoTube Starfooted Cryogenic Vials with intern (379189).
More Details
Monitors Panel
Buy Monitors Panel Items Including:
CALgrafix Process Monitoring and Configuration Software (other license
CALgrafix Process Monitoring and Configuration Software (Add a license (1005GB000)
Trace Air OV/IAQ Badge with PrePaid Analysis PK/5.
More Details
Delrin Quick-Disconnect
Buy Delrin Quick-Disconnect Items Including:
Miniature Quick-Disconnect Coupling; Acetal; Straight-through, 1/8" ID (SMC02)
Quick-disconnect fittings, Standard John Guest valved Fittings, Delri (PLCD14004)
Quick-disconnect fittings, Standard John Guest valved fittings, Delrin (PLCD14006).
More Details
Mixing Tubes
Buy Mixing Tubes Items Including:
Pump and mix tube set, 39", Stainless Steel (F426S 41/38 39")
Mixing drum pump, 24 GPM, PP, sealless pump tube
Cole-Parmer Aluminum Block, for 40 x 0.5-mL Microcentrifuge Tubes
Cole-Parmer Aluminum Blocks, for 12 x 15/16mm ID Test Tubes.
More Details
Cole-Parmer Centrifuges
Buy Cole-Parmer Centrifuges Items Including:
Cole-Parmer Centrifuge, variable speed, 115 VAC, 60 Hz
Cole-Parmer Personal Microcentrifuge; 115V; 60 Hz (YO-17307-00)
Cole-Parmer Personal Microcentrifuge; 230V; 50Hz (YO-17307-05)
Cole-Parmer StableTemp Single Block Combination, Centrifuge Tubes.
More Details
Magnehelic Gauges
Buy Magnehelic Gauges Items Including:
Mounting Kit for Magnehelic Gauges
Air Filter Kit for Magnehelic Gauges
Adjustable Signal flag for Magnehelic Gauges
Dwyer Magnehelic Differential Pressure Gauge, 2002: 0-2.0" WC
Dwyer Magnehelic Differential Pressure Gauge, 2040: 0/40 Inh2O.
More Details
Thermocouple Monitor
Buy Thermocouple Monitor Items Including:
Cole-Parmer Remote-Monitoring Thermocouple Thermometer (Deg
F)
Cole-Parmer Remote-Monitoring Thermocouple Thermometer (Deg
C).
More Details
Stainless Steel Braided
Buy Stainless Steel Braided Items Including:
Stainless Steel Braided Chemfluor PTFE Hose; 13 ft L; 1/8" ID
Stainless Steel Braided Smooth Chemfluor PTFE Hose with Silicone Cover (YO-95645-00)
Stainless Steel Braided Smooth Chemfluor PTFE Hose with Silicone Cover (YO-95646-00).
More Details
Electric Meter Calibration
Buy Electric Meter Calibration Items Including:
NIST-Traceable Calibration; Watt Meter (YO-17090-28)
NIST-Traceable Calibration; Power Quality Meter (YO-17090-34)
NIST-Traceable Calibration; Capacitance Meter (YO-17090-38)
NIST-Traceable Calibration; Conductivity/Resistivity Meter (YO-17090-30).
More Details
Colder Quick-Disconnect Inserts (Male Coupling Halves)
Buy Colder Quick-Disconnect Inserts (Male Coupling Halves) Items Including:
Quick-disconnect fittings, Straight-through Male pipe thread inserts, (HFC241212)
Quick-disconnect fittings, Straight-through hose barbs inserts, PP, 3/ (HFC221012)
Quick-disconnect fittings, Straight-through hose barbs inserts, PP, 3/ (HFC221212).
More Details
Ohaus Weights
Buy Ohaus Weights Items Including:
500 g Attachment Weight for Ohaus Mechanical Scales
1000 g Attachment Weight for Ohaus Mechanical Scales
Ohaus Triple Pro Mechanical Balance, 2610g X 0.1g w/attachment weights.
More Details
Disposable Test Tubes
Buy Disposable Test Tubes Items Including:
Kimble Disposable Culture Tubes, 13 x 100, with markings 1000/cs
Kimble Disposable Culture Tubes, 16 x 100, with markings 1000/cs
Kimble Disposable Culture Tubes, 16 x 125, with markings 1000/cs
Disposable polypropylene test/culture tubes, 20 mL.
More Details
Cooling Range
Buy Cooling Range Items Including:
Cole-Parmer® Polystat® CC1080 SEALED CIRCULATOR 120V (YO-12123-50).
More Details
Fluke T5 600
Buy Fluke T5 600 Items Including:
Fluke T5-600/62MAX+/1AC Electrical/IR Kit
Fluke T5-600/62/1AC: II, IR Thermometer/Electrical Combo Kit.
More Details
Load Cell Systems
Buy Load Cell Systems Items Including:
22 lb capacity smart "S" type load cell
44 lb capacity smart "S" type load cell
110 lb capacity smart "S" type load cell.
More Details
Fifty Gallon Drum
Buy Fifty Gallon Drum Items Including:
Fifty-five gallon drum heater, 230 VAC (309-55GAL)
Fifty-five gallon drum adapter kit for 13- and 44-GPM models
Fifty-five gallon drum adapter kit
Dolly Heated 55gal.drum 120v (HDD-55/120V).
More Details
Cole-Parmer Hollow Fiber Cartridge Filters
Buy Cole-Parmer Hollow Fiber Cartridge Filters Items Including:
Hollow fiber cartridge filter; 0.1 µm, 10"L
Hollow fiber cartridge filter; 0.1 µm, 20"L
Hollow fiber cartridge filter; 0.2 µm, 20"L.
More Details
Stainless Steel Filters
Buy Stainless Steel Filters Items Including:
Advantec Stainless Steel Filter Holder, Syringe; 13mm dia
Advantec Stainless Steel Filter Holder, Syringe; 25mm dia
Advantec High-Pressure Stainless Steel Filter Holder, 47 mm
Advantec All-Stainless Steel Filter Holder for 47-mm Membranes, Stainl.
More Details
Cole-Parmer 316 Ss Fittings
Buy Cole-Parmer 316 Ss Fittings Items Including:
316 SS fittings; male slip plug (YO-31507-22)
316 SS fittings; male luer lock plug (YO-31507-23)
316 SS fittings; female luer x 1/8" hose barb (YO-31507-28)
Luer fittings, 316 SS fittings, female luer lock x 1/16" hose barb (YO-31507-29).
More Details
Gast Vacuum Pumps
Buy Gast Vacuum Pumps Items Including:
Vacuum/pressure diaphragm pump with gauges, regulators, and relief val (DAA-V515-ED)
Vacuum/pressure diaphragm pump, single head, 1.1 cfm, 220/240VAC (DOA-P725-BN)
Vacuum/pressure diaphragm pump with gauges, regulators, and relief val (DOA-P704-AA).
More Details
Rms Meters
Buy Rms Meters Items Including:
Extech 380976-K 1 and 3 Phase 1000A TRMS Power Clamp Meter
Panel Meter With Rms Volt And Current Inputs; Green Lcd
502 : True Rms Clamp Meter, 400A AC/DC 600V AC/DC (502)
2129.53 : Clamp-On Meter True RMS 400AAC/DC 600VAC/DC Hz Power (Kw) Ph (F05).
More Details
Carbon Filter Cartridge
Buy Carbon Filter Cartridge Items Including:
Double open end high-efficiency pleated polypropylene filter cartridge (P0.2-10E)
Double open end pleated carbon-impregnated cartridge; 10 µm, 10"L (155367-43)
Double open end pleated carbon-impregnated cartridge; 10 µm, 20"L (155397-43).
More Details
Silicone Rubber Cord
Buy Silicone Rubber Cord Items Including:
All-purpose silicone rubber heat mat, 10" L x 2" W, 100 watts, 120 VAC (020100C1)
All-purpose silicone rubber heat mat, 10" L x 3" W, 150 watts, 120 VAC (030100C1).
More Details
High Temperature Thermistor
Buy High Temperature Thermistor Items Including:
Fluke Calibration (formerly Hart Scientific) Probe1000 C10 Ohm Prt 0.2 (5624-20-B)
Fluke Calibration (formerly Hart Scientific) Probe1000C10 Ohm Prt 0.25 (5624-20-D)
Fluke Calibration (formerly Hart Scientific) Probe1000 C10 Ohm Prt 0.2 (5624-20-G).
More Details
Colder Npt(M) Threaded Adapter Inserts
Buy Colder Npt(M) Threaded Adapter Inserts Items Including:
Quick-Disconnect NPT(M) Pipe Adapter Insert; Straight-through, Delrin, (PLC24004)
Quick-Disconnect NPT(M) Pipe Adapter Insert; Straight-through, PP, 1/8 (PMC240212)
Quick-Disconnect NPT(M) Pipe Adapter Insert; Straight-through, PP, 1/4 (PLC2400412).
More Details
70618
results
<<<Previous
13
14
15
16
17
18
19
20
21
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
40
50
60
70
80
90
100
110
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Hvac to Colder Npt(M) Threaded Adapter Inserts
>
31125329-s-toc_2 31125329stoc2