Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 2
Cole-Parmer Burettes (Burets)
to
Cole-Parmer Static (in-Line) Mixers
Image
Product Detail
More Detail
Cole-Parmer Burettes (Burets)
Buy Cole-Parmer Burettes (Burets) Items Including:
COLE-PARMER BURETTE, 10ML WITH GLASS STOPCOCK
COLE PARMER BURETTE, 25ML WITH GLASS STOPCOCK
COLE PARMER BURETTE, 100ML WITH GLASS STOPCOCK
COLE PARMER BURETTE, 10ML WITH GLASS STOPCOCK, 6/CASE
COLE PARMER BURETTE, 25ML WITH GLASS STOPCOCK, 6/CASE.
More Details
Water Metering Pump
Buy Water Metering Pump Items Including:
Etatron HDS pH Pump Control System, 5 L/hr, 10 bar max pressure, wall-
Etatron HDS pH Pump Control System, 10 L/hr, 10 bar max pressure, wall
Etatron DLX pH/ORP Pump Control System, 1 L/hr, 15 bar max pressure, w (DLX-pH-RX/MBB) (YO-56022-40).
More Details
Marathon Microscopes & Magnification
Buy Marathon Microscopes & Magnification Items Including:
8 x 40 Binoculars with Built-In Sun Filter (BI-030014).
More Details
Cole-Parmer Coiled Fep Tubing
Buy Cole-Parmer Coiled Fep Tubing Items Including:
Coiled FEP tubing, 1/8"ID x 1/4"OD, 2 ft long (YO-95821-00)
Coiled FEP tubing, 1/8"ID x 1/4"OD, 4 ft long (YO-95821-01)
Coiled FEP tubing, 1/4"ID x 3/8"OD, 2 ft long (YO-95821-02).
More Details
Industrial Shop Vacuums
Buy Industrial Shop Vacuums Items Including:
Koehler Manual Vacuum Distillation System, 220-240 VAC, 50/60 Hz
Koehler Vacuum System for cold filter plugging point apparatus
100/5000PSI Oil Operated Single Piston
100/10000PSI Oil Operated Single Piston
100/16000PSI Oil Operated Single Piston.
More Details
Tube 42
Buy Tube 42 Items Including:
Microbore - Puri-Flex ™ Autoanalysis Tubing, -42, 50 feet
3-Stop Tygon® Lab Tubing, 2.06 mm ID, 12/pack (CP95608-42)
Foam Test Tube Rack-small, 6 per pack
Horizontal Tube Furnace, 42-1/8" Tube Length, 1500°C, 208 / 240 V (RHTC 80-710/15/P330).
More Details
Low Speed Motors
Buy Low Speed Motors Items Including:
Shimpo FGS-100E-L Test Stand, 110 lb, Low Speed, 100 to 230 VAC
Low speed test stand
Motorized Force Gauge Test Stand, Vertical low speed (YO-93950-92).
More Details
Parker Hannifin Bulkhead Union Fittings
Buy Parker Hannifin Bulkhead Union Fittings Items Including:
Compression fittings, Bulkhead unions; Black PP, 3/8" OD, 2-5/8" x 15/ (YO-06444-32)
Compression bulkhead union, PFA, 1/8 (YO-31320-82)
Compression bulkhead union, PFA, 1/4 (YO-31320-83)
Compression bulkhead union, PFA, 3/8 (YO-31320-84).
More Details
Air Compressor Air Lines
Buy Air Compressor Air Lines Items Including:
Air Tool, Straight Line Sander, 16 in x 2-1/2 in pad, 2,500 rpm, 1/4 i (AT-7019)
Quick Disconnect Air Coupler, 1/4 in NPT(F) (MS-1/4F)
Quick Disconnect Air Coupler, 3/8 in NPT(F) (MS-3/8F)
Quick Disconnect Air Coupler, 1/4 in NPT(M) (MS-1/4M).
More Details
Microscope Eyepieces
Buy Microscope Eyepieces Items Including:
Replacement eyepiece with reticle for 03890-10, 10x magnification (YO-03890-51)
Replacement eyepiece with reticle for 03890-20, 10x magnification (YO-03890-52)
Eyepiece without reticle, 10X (YO-03890-55)
Eyepieces, 15 x magnification (YO-03904-50).
More Details
Labels and Labeling
Buy Labels and Labeling Items Including:
Dymo LabelWriter 450, Uses One Label Writer Label
Dymo LabelWriter 450 Duo, Uses One Label Writer Label and One D1 Label
Address Label, 130 Labels Per Roll, Two Roll/Pack
Waterproof label, 2"W x 1"H
Waterproof label, 4"W x 2"H.
More Details
Air Control Valves
Buy Air Control Valves Items Including:
High-Pressure Flow Control Valve
2-Position, 3-Way Body Ported Valve; 0-120 psig; 120VAC (XM30NBG53A)
2-Position, 3-Way Body Ported Valve; -14.7 to 15psig; 24VDC (XM30VBG49A)
2-Position, 3-Way Body Ported Valve; -14.7 to 15psig; 12VDC (XM30VBG45A).
More Details
Arc Flash Requirements
Buy Arc Flash Requirements Items Including:
Arc Flash and Shock Hazard Appropriate PPE Required label, 5 x 7, ea (LELC315)
Warning Arc Flash and Shock Hazard Appropriate PPE Required, 5" x 7" (LELC325)
Warning Arc Flash and Shock Hazard Appropriate PPE Required When Worki (LELC309).
More Details
Heated Cup
Buy Heated Cup Items Including:
Pensky-Martens gas-heated closed-cup flash tester
Koehler tag gas heated closed-cup flash tester.
More Details
Scienceware Labware
Buy Scienceware Labware Items Including:
Scienceware Separatory Funnel Rack
Steel forceps with PTFE coating (37937)
Stainless steel tongs with PTFE coated tips (37945).
More Details
Cole-Parmer Black (Uv) Fluorescent Ring Lights
Buy Cole-Parmer Black (Uv) Fluorescent Ring Lights Items Including:
Black (UV) light ring lights, for 48402-series stereomicroscopes; 115V (YO-41210-00)
Black (UV) light ring lights, for 48403-series stereomicroscopes; 115V (YO-41210-10).
More Details
Chempette Dispensers
Buy Chempette Dispensers Items Including:
Chempette Compact Dispenser -0.2-1mL (A3-AJ-1)
Chempette Compact Dispenser - 0.4-2mL (A3-AJ-2)
Chempette Compact Dispenser 1-5mL (A3-AJ-5)
Chempette Compact Dispenser 2-10mL (A3-AJ-10)
Chempette digital repetitive dispenser, 0.2-1 mL (A3-21DG-1).
More Details
Koehler Instruments
Buy Koehler Instruments Items Including:
Eutech Instruments pH 200 1/8-DIN pH/ORP controller
Eutech Instruments pH 1000 controller, 115 VAC
Eutech Instruments pH 1000 controller, 220 VAC
Eutech Instruments pH 2000D, differential pH/ORP Controller
Eutech Instruments pH 550 pH/orp Monitor.
More Details
Cole-Parmer Glassware for Vapodest Distillation Systems
Buy Cole-Parmer Glassware for Vapodest Distillation Systems Items Including:
Glassware for Vapodest Distillation Systems, 250mL Kjeldahl flask.
More Details
Digi-Sense Industrial Thermocouple Probes
Buy Digi-Sense Industrial Thermocouple Probes Items Including:
Type J Industrial Direct Insert Ungrounded Thermocouple Probe 12"L (700-J-F-12-W4-316-AL-1/2)
Type K Industrial Direct Insert Ungrounded Thermocouple Probe 12"L (700-K-F-12-W4-316-AL-1/2).
More Details
Pneumatic Calibration
Buy Pneumatic Calibration Items Including:
Pneumatic Test Pump -13/600psi
NIST-Traceable Calibration; Pneumatic Deadweight Tester, 0.1% (YO-17101-83)
NIST-Traceable Calibration; Pneumatic Deadweight Tester, 0.015% (YO-17101-85)
Pneumatic Pressure and Vacuum Pump, 0-600 PSI; 0-28.5" Hg.
More Details
Bioreactor Vessel
Buy Bioreactor Vessel Items Including:
Reaction vessel, 3 oz
Reaction vessel, 6 oz
Reaction vessel, 12 oz
Reaction vessel with clear plastic shield and coupling, 3 oz
Reaction vessel with clear plastic shield and coupling, 12 oz
More Details
Seward Reciprocating Paddle Blenders
Buy Seward Reciprocating Paddle Blenders Items Including:
Seward Stomacher 80 Lab Blenders, compact, 110VAC (YO-36850-00)
Seward Stomacher 80 Lab Blenders, compact, 230VAC (YO-36850-02)
Seward Stomacher 400C Lab Blenders, programmable, 110VAC (YO-36850-04).
More Details
Field Notebooks
Buy Field Notebooks Items Including:
Thermo Scientific Nalgene field notebook
4-5/8" x 7" spiral notebook - field pattern (353N-6).
More Details
Hazardous Duty Mixers
Buy Hazardous Duty Mixers Items Including:
Low-cost, Direct-drive Air Motor Mixer; 200-10,000 RPM (YO-04680-00)
Low-cost, gear-drive air motor mixer with steel gears; 50-1200 rpm (YO-04681-10)
Low-cost, Direct-drive Air Motor Mixer; 200-7,000 RPM (YO-04686-00).
More Details
Bottle Cap Torque
Buy Bottle Cap Torque Items Including:
Mark-10 MR53-20Z Torque Sensor, 20 oz x 1400 g cm x 14 N cm
Mark-10 MR53-12 Torque Sensor, 12 lb x 192 oz in x 135 N cm
Mark-10 MR53-50 Torque Sensor, 50 lb x 800 oz in x 570 N cm
Mark-10 MR53-100 Torque Sensor, 100 lb x 1600 oz in x 1150 N cm.
More Details
Thermometer with Clock
Buy Thermometer with Clock Items Including:
Cole-Parmer Wall-Mount Digital Clock/Calendar/Thermometer
Cole-Parmer Thermometer/Clock/Humidity Thermometer
Wall Clock / Hygro-Thermometer Radio Controlled with Alarm (7166CC)
Extech CTH10 Radio Controlled Clock Hygro-Thermometer.
More Details
Silastic Tubing
Buy Silastic Tubing Items Including:
Silastic Laboratory Tubing, .012" ID x .025" OD, 50'.
More Details
Tissue Culture
Buy Tissue Culture Items Including:
Cole-Parmer Sterile Tissue Culture 6-Well Plate, 100/case
Cole-Parmer Sterile Tissue Culture 12- Well Plate, 100/case
Cole-Parmer Sterile Tissue Culture 24-Well Plate, 100/case
Cole-Parmer Sterile Non-Treated Tissue Culture 96-Well Plate, 100/case.
More Details
Cole-Parmer Static (in-Line) Mixers
Buy Cole-Parmer Static (in-Line) Mixers Items Including:
Mounted in-line mixer; 5/16" tube OD, 12 elements, 10/pack (YO-04668-02)
Mounted in-line mixer; 3/8" tube OD, 12 elements, 10/pack (YO-04668-04)
Mounted in-line mixer; 1/2" tube OD, 12 elements, 10/pack (YO-04668-06).
More Details
70618
results
<<<Previous
1
2
3
4
5
6
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Cole-Parmer Burettes (Burets) to Cole-Parmer Static (in-Line) Mixers
>
31125329-s-toc_2 31125329stoc2