Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 200

Gallon Dispenser   to   K Resin

Image
  Product Detail
More Detail

Gallon Dispenser

Buy Gallon Dispenser Items Including:
  • DrumQuik PUR Drum Dip Tube for 5-gallon chem pack, 334 mm length, HDPE (YO-31608-72)
  • DrumQuik PUR Drum Dip Tube for 55-gallon drum, 985 mm length, HDPE, pa (YO-31608-73)
  • Paraffin Dispenser, microprocessor controlled, 6.25 gallon capacity w/.

Cole-Parmer Hand-Held Single-Grip Mixers

Buy Cole-Parmer Hand-Held Single-Grip Mixers Items Including:
  • Hand-held single-grip mixer; 1/4" dia shaft x 15"L, 2-1/2" dia caged i (YO-50500-45)
  • Hand-held single-grip mixer; 1/2" dia shaft x 21"L, 5" dia caged impel (YO-50500-47).

Digital Vacuum Controller

Buy Digital Vacuum Controller Items Including:
  • Digital Pressure Controller, 0 To 15 PSI ~ 3/4" NPT(F)
  • Digital Pressure Controller, 0 To 30 PSI ~ 3/4" NPT(F)
  • Digital Pressure Controller, 0 To 100 PSI ~ 3/4" NPT(F)
  • Digital Pressure Controller, 0 To 15 PSI ~ 1/8" NPT(F).

Water Sampling

Buy Water Sampling Items Including:
  • Whirl-Pak Coli-Test sodium thiosulfate bags for water sampling, 4 oz
  • Whirl-Pak sodium thiosulfate bags for potable water sampling, 4 oz
  • Global Water hydrant water pressure logger (YO-32922-82)
  • Global Water garden hose pressure logger (YO-32922-70).

Nalgene Safety Products

Buy Nalgene Safety Products Items Including:
  • Thermo Scientific Nalgene Fluorinated HDPE Safety Waste System
  • Thermo Scientific Nalgene "Right-to-Know" safety wash bottle for ethan (2425-0502)
  • Thermo Scientific Nalgene "Right-to-Know" safety wash bottle for ethan (2425-1002).

Cole-Parmer Replacement Probes

Buy Cole-Parmer Replacement Probes Items Including:
  • Coupler for Cole-Parmer 500- and 750-Watt Processors (630-0421)
  • Micro Probe for Cole-Parmer Scientic Thermistor Thermometer 90080-12 (YO-90080-14)
  • Replacement Probe for Thermometer 90080-00
  • Probe, 2-mm (5/64 (630-0423)
  • Probe, 3-mm (1/8 (630-0422).

Temperature Rh Logger

Buy Temperature Rh Logger Items Including:
  • MicroLite II Temperature/RH USB Data Logger
  • Simple Logger II Temperature/RH Model L702.

Air Powered Mixers

Buy Air Powered Mixers Items Including:
  • Lightnin general-purpose air-drive mixer
  • Low-cost, gear-drive air motor mixer; 8-170 rpm (YO-04681-00)
  • Low-cost, gear-drive air motor mixer with steel gears; 50-1200 rpm (YO-04681-10)
  • Low-cost, gear-drive air motor mixer; 50-1200 rpm (YO-04685-00).

Cole-Parmer Humidity Psychrometers / Mason Hygrometers

Buy Cole-Parmer Humidity Psychrometers / Mason Hygrometers Items Including:
  • Mason Hygrometer (7109)
  • Cole-Parmer Psychrometer, F (22014 ColeParmer labeled)
  • Cole-Parmer Psychrometer, C (22000 ColeParmer labeled)
  • Humidity Slide Calculator (HC880).

Cole-Parmer Cellulose Acetate

Buy Cole-Parmer Cellulose Acetate Items Including:
  • Cole-Parmer Cellulose Acetate Sterile Filters; pore size; 0.45µ; Ac
  • Cole-Parmer HPLC Syringe Filters, Non Sterile, Cellulose Acetate (CA), (YO-32817-14)
  • Cole-Parmer HPLC Syringe Filters, Non Sterile, Cellulose Acetate (CA), (YO-32817-16).

Static Mixers

Buy Static Mixers Items Including:
  • Standard gear pump system with static mixer and 0.5 to 1.0 GPM flow ra
  • Standard gear pump system with static mixer and 3.0 to 7.0 GPM flow ra
  • Standard gear pump system with calibration column, static mixer, and 0.

Switches and Relays

Buy Switches and Relays Items Including:
  • Flowline Switch-Pro Remote Relay Level Controller; one sensor, one rel
  • Flowline Switch-Pro Remote Relay Level Controller; two sensors, one re
  • Cole-Parmer® programmable process meter with two SPDT relays, 115 (YO-94785-10).

Pyrex Analytical Funnels

Buy Pyrex Analytical Funnels Items Including:
  • Pyrex Brand 6180 funnel; 50 mm top dia, 108 mm L, pack of 12
  • Pyrex Brand 6180 funnel; 65 mm top dia, 121 mm L, pack of 12
  • Pyrex Brand 6180 funnel; 75 mm top dia, 139 mm L, pack of 12
  • Pyrex Brand 6180 funnel; 100 mm top dia, 190 mm L, pack of 6.

Parker Microbiology

Buy Parker Microbiology Items Including:
  • Contact plate, Baird Parker Agar, 10/pk
  • Prepared agar plate, Baird Parker agar, 10/pk
  • Cole-Parmer Baird Parker Agar, 500 g
  • Baird Parker Agar Base 500g.

Uv Sensors

Buy Uv Sensors Items Including:
  • UV-B sensor for single-input datalogging radiometer (PMA2101)
  • UV Safety Sensor for Measuring Efeective Actinic UV Irradiance (PMA2120)
  • UV-C Germicidal Sensor for Dual-input datalogging or Singel-input Radi (PMA2122)
  • UV sensor, 312 nm (YO-97503-52).

Digital Industrial Scales

Buy Digital Industrial Scales Items Including:
  • Reichert r2 mini Digital Pocket Refractometer, Brix and Ref
  • Atago Programmable Digital Refractometer, ±0.1% Brix accuracy
  • Atago Programmable Digital Refractometer, ±0.03% Brix accuracy and
  • SS, 0-24", large scale digital caliper (54-100-024-1).

Rs232 Sub

Buy Rs232 Sub Items Including:
  • 2 Meter Communication Cable (RS232 - 9 Pin D-Sub) for Pump to Pump use (788308) (YO-74903-77).

Erie Scientific Co. Clipped-Corner Slides

Buy Erie Scientific Co. Clipped-Corner Slides Items Including:
  • Superfrost Clipped-Corner Slides, 144/PK (4951-4-001).

Deionizer Cartridge

Buy Deionizer Cartridge Items Including:
  • Thermo Scientific Barnstead Ultrapure deionization cartridge, 680 grai (D50253)
  • Ultra-low organics deionized feed for Thermo Scientific Barnstead EASY (D502124)
  • Thermo Scientific Barnstead Pretreatment Cartridge for tap and deioniz (D50251).

Potentiometric Titration

Buy Potentiometric Titration Items Including:
  • Schott manual volumetric titrator, 20-mL capacity, 115 VAC
  • Schott automatic volumetric titrator, 20-mL capacity, 115 VAC
  • Schott automatic volumetric titrator, 20-mL capacity, 230 VAC
  • Mettler Toledo Easy pH Titrator
  • Mettler Toledo Easy Ox Titrator.

Bw Technologies

Buy Bw Technologies Items Including:
  • BW Technologies Microdock II Base Station with GasAlertClip Extreme Ca
  • BW Technologies Microdock II Base Station with GasAlertMicro 5 module (YO-86419-59)
  • BW Technologies Microdock II Base Station with GasAlertMicro 5 module (YO-86419-60).

Ph Treatment

Buy Ph Treatment Items Including:
  • Fill solution for chlorine dioxide sensors, 30-mL bottle (YO-99673-11)
  • Fill solution for Free Chlorine sensors, 30-mL bottle (YO-99673-13)
  • Replacement membrane cap for Chlorine Dioxide sensors (YO-99673-15).

Fine Tip Permanent Markers

Buy Fine Tip Permanent Markers Items Including:
  • Fine-tip scientific marking pen, black (YO-09965-10)
  • Fine-tip scientific marking pen, blue (YO-09965-30)
  • Fine-tip scientific marking pen, green (YO-09965-40)
  • Sharpie™ industrial marker with fine tip, black (YO-09969-00).

Parker Hannifin Straight Unions

Buy Parker Hannifin Straight Unions Items Including:
  • Push-to-connect Straight Union Connector, PVDF, 1/4" Tube OD, Pack Of (F4UC4)
  • Push-to-connect Straight Union Connector, PVDF, 3/8" Tube OD, Pack Of (F6UC6)
  • Compression Union, 5/16" x 5/16", Stainless Steel, Each (YO-30486-02).

Hermle Labnet Centrifuges

Buy Hermle Labnet Centrifuges Items Including:
  • Hermle High-Capacity Refrigerated Microcentrifuge; 120VAC, 50/60Hz
  • Microcentrifuge Rotor, hermetically sealed; 24 position, 1.5/2.0mL tub (YO-17405-62)
  • Microcentrifuge Rotor, hermetically sealed; 30 position, 1.5/2.0mL tub (YO-17405-63).

Cole-Parmer Acetic Acid, Sodium Salt

Buy Cole-Parmer Acetic Acid, Sodium Salt Items Including:
  • ACETIC ACID SODIUM SALT 500GR (YO-88362-67).

Biospec Products

Buy Biospec Products Items Including:
  • Torbeo Hand-Held Homogenizer; capacity 1 to 200 mL; stator size; 1.3 c (YO-04751-05)
  • Torbeo Hand-Held Homogenizer; capacity 100 to 1000 mL; stator size; 2
  • (YO-04751-10)
  • Thermal Printer Kit for the UVmini-1240 and BioSpec-mini Spectrophotom.

Hand Pumps for Oil

Buy Hand Pumps for Oil Items Including:
  • Pump, Motorized Drum Pump Package, Designed for Light Oils, 35 GPM, 31 (9410-110V) (YO-70614-10)
  • Pump, Motorized Drum Pump Package, Designed for Light Oils, 35 GPM, 31 (9411-230V) (YO-70614-11).

Uv Light Detector

Buy Uv Light Detector Items Including:
  • UVX Digital UV Intensity Meter (97-0015-02(UVX))
  • UV Intensity Meter Sensor, 365 nm (97-0016-02(UVX36))
  • UV Intensity Meter Sensor, 302 nm (97-0016-04(UVX31)).

K Resin

Buy K Resin Items Including:
  • Cole-Parmer® Fluorimeter cuvettes with cover of PTFE resin; glass; (YO-83303-02)
  • Cole-Parmer® Fluorimeter cuvettes with cover of PTFE resin; glass; (YO-83303-03)
  • Cole-Parmer® Fluorimeter cuvettes with cover of PTFE resin; glass; (YO-83303-04).



Gallon Dispenser to K Resin

>