Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 21
Goggles Safety Uvex
to
6 Port Manifold
Image
Product Detail
More Detail
Goggles Safety Uvex
Buy Goggles Safety Uvex Items Including:
Uvex STEALTH Safety Goggles, Clear Lens
Uvex Strategy Safety Goggles with Fabric Band, Gray Body with Direct V
Uvex Strategy Safety Goggles with Neoprene Band, Gray Body with Indire
Flex Seal Safety Goggle, gray lens (S3410X)
Laser Goggles, HE-NE.
More Details
Cole-Parmer Palladium
Buy Cole-Parmer Palladium Items Including:
PALLADIUM ON BARIUM SULFAT 10G (YO-88289-58).
More Details
Filter 15
Buy Filter 15 Items Including:
Ks13 PTFE O-ring (YO-02930-15)
Whatman Quantitative Filter Papers-Ashless less than 0.010% ash; 15 cm (YO-06647-15)
Whatman Qualitative Filter Papers; 15 cm dia; pore size, 11 µ; 100/
Whatman™ Klari-Flex™ Filter Funnel; 0.45 um PES, 1000mL, 12/PK.
More Details
Moisture Analyzer
Buy Moisture Analyzer Items Including:
Printer, for Mettler Toledo moisture analyzers 11334-50 and -51
Printer, for Mettler Toledo moisture analyzer 11334-52
GE (General Eastern) Industrial relative Humidity/Dew Point Analyzer, (MR2350) (YO-37405-04)
Universal Thermohygrometer (4080CP).
More Details
Bsp Fittings
Buy Bsp Fittings Items Including:
Pipe fittings, NPT(F) x BSPT(M) adapter, BR, 1/4", 6200 psi, 1-3/8", (YO-31834-01)
Coupling Body, Non-Spill, Threaded, 3/8" male BSP (NSHD10006BSPT)
Coupling Insert, Non-Spill, Threaded, 3/8" male BSP (NSHD24006BSPT).
More Details
Ametek Calibrators
Buy Ametek Calibrators Items Including:
Pressure Module,-15-200psi.
More Details
Dry Baths
Buy Dry Baths Items Including:
Beads for dry bath, 0.75-liter
Fluke Calibration (formerly Hart Scientific) 9103-A:Low Temp
Dry-Well
Fluke Calibration (formerly Hart Scientific) 9140-A:Dry-Well Calibrato
Cole-Parmer StableTemp Stainless Steel Sand Baths, 1 block models.
More Details
Gradient Pcr
Buy Gradient Pcr Items Including:
3 Prime X USB Gradient Upgrade
Techne® TC-4000 Standard Non-Gradient Thermocycler with 96 x 0.2ml (YO-93943-01)
Techne® TC-4000 Standard Non-Gradient Thermocycler with 60 x 0.5ml
Techne® TC-4000 Standard Non-Gradient Thermocycler with 384-well p.
More Details
Whatman Laboratory Filtration Products
Buy Whatman Laboratory Filtration Products Items Including:
Whatman Quantitative Filter Papers-Ashless less than 0.010% ash; 5.5 c (YO-06647-01)
Whatman Quantitative Filter Papers-Ashless less than 0.010% ash; 9.0 c (YO-06647-02).
More Details
Lab Products Sterilized Sampling Bottle Assemblies
Buy Lab Products Sterilized Sampling Bottle Assemblies Items Including:
Sterilized Sampling Bottle Assembly, 125 mL
Sterilized Sampling Bottle Assembly, 250 mL
Sterilized Sampling Bottle Assembly, 500 mL
Sterilized Sampling Bottle Assembly, 1 L.
More Details
Force Sensors
Buy Force Sensors Items Including:
Mark-10 MR02-1000 Force Sensor, 1000 lb x 500 kg x 5000 N
Mark-10 MR02-2000 Force Sensor, 2000 lb x 1000 kg x 10000 N
Mark-10 MR02-5000 Force Sensor, 5000 lb x 2500 kg x 25000 N
Mark-10 MR01-2000 Force Sensor, 2000 lb x 1000 kg x 10000 N.
More Details
Dwyer Inclined Manometer
Buy Dwyer Inclined Manometer Items Including:
200.5: Inclin Mnmtr.1/0/1inW Durablock Stat Inclined Manometer 10/0/1i
209:Incline Mnmtr.2/0/3 Inw Durablock Stat Inclined Manometer .2/0/3.0
202.5:Inclin Mnmtr.2/0/2inW Durablock Stat Inclined Mnometer 0.2/0/2.0
Red Gauge Oil 4 oz.
More Details
Thuemling Pressure & Vacuum Instruments
Buy Thuemling Pressure & Vacuum Instruments Items Including:
SPAN Pressure Gauge, Vac to 30 psi (YO-68081-02)
SPAN Pressure Gauge, 0 to 5000 psi (YO-68081-28)
SPAN Pressure Gauge, 0 to 100 psi (YO-68081-40)
SPAN Pressure Gauge, 0 to 1000 psi (YO-68081-52)
SPAN Pressure Gauge, 0 to 2000 psi (YO-68081-54).
More Details
Tds Calibration
Buy Tds Calibration Items Including:
Oakton TDS 6+ handheld TDS meter kit with NIST-traceable Calibration.
More Details
Digital Titrator
Buy Digital Titrator Items Including:
Hach digital titrator (16900-01)
Alkalinity titrator cartridge with range of 10 to 160 mg/L CaCO3 and t (14388-01)
Chloride titrator cartridge with range of 2.5 to 150 g/L C-; 100 to 10 (14397-01)
Hirschmann opus Titrator, 0.1 to 200 mL, 120 VAC.
More Details
Horiba U-10
Buy Horiba U-10 Items Including:
Replacement DO sensor for Horiba U-10 series meters (362174)
Horiba B-171 Twin Conductivity/Salinity Pocket Tester
Horiba Laqua Tester, pH
Horiba U-52G Multiparameter Meter, 33-ft (10-m) cable
Horiba U-53G Multiparameter Meter, 33-ft (10-m) cable.
More Details
Ph Water Treatment
Buy Ph Water Treatment Items Including:
Fill solution for chlorine dioxide sensors, 30-mL bottle (YO-99673-11)
Fill solution for Free Chlorine sensors, 30-mL bottle (YO-99673-13)
Replacement membrane cap for Chlorine Dioxide sensors (YO-99673-15).
More Details
Cole-Parmer Pressure & Vacuum Controllers
Buy Cole-Parmer Pressure & Vacuum Controllers Items Including:
1 to 1500 torr Cole-Parmer Pressure/Vacuum Pirani-Type Sensor, 1/4" NP
30" Hg To 100 PSI Cole-parmer Pressure/vacuum Diaphragm-type Sensor, 1
0 to 200 psi Cole-Parmer Pressure/Vacuum Diaphragm-Type Sensor, 1/4" N.
More Details
Vortex Genie
Buy Vortex Genie Items Including:
Double-tier 96-well Microwell Plate Insert for Vortex-Genie 2™ (SI-0510)
Replacement Cup Head for Vortex-Genie 2™ (146-3011-00)
Replacement Elastic Bands for Vortex-Genie 2™, 2/set (568-0001-00)
Small ampule attachment for Vortex Genie 2 (SI-0570).
More Details
Degassing Water
Buy Degassing Water Items Including:
Sample Filtration and Degassing Kit (YO-99511-16).
More Details
Techne Fluidized Bath
Buy Techne Fluidized Bath Items Including:
Fluidized sand bath systems; temperature range; 50-600°C; 117V; 2x (YO-01184-00)
Fluidized sand bath systems; temperature range; 50-600°C; 240V; 2x (YO-01184-02)
Fluidized sand bath systems; temperature range; 50-600°C; 240V; 4x (YO-01189-50).
More Details
Eyewash Stations
Buy Eyewash Stations Items Including:
Universal cart, mount and transport most portable eyewash stations (32-001060-0000)
Fend-all Primary Eyewash Station (32-000200-0000)
Honeywell100% Sterile Primary Eyewash Station with Alarm
Scienceware Plum Sterile Saline Eyewash Station, 1000 mL.
More Details
Ptfe Coatings
Buy Ptfe Coatings Items Including:
Vacuum/pressure Diaphragm Pump With Gauges And Regulators, PTFE-coated (107CA18TFEL-246)
Vacuum/pressure diaphragm pump with gauges and regulators, PTFE-coated (107CD18TFEL-246)
Vacuum/pressure diaphragm pumps, PTFE-coated wetted parts, 0.75 cfm, 1.
More Details
Autoclave Tray
Buy Autoclave Tray Items Including:
Thermo Scientific Nalgene polypropylene autoclaving tray, 1 L
Thermo Scientific Nalgene polypropylene autoclaving tray, 2 L
Thermo Scientific Nalgene polypropylene autoclaving tray, 3 L
Thermo Scientific Nalgene polypropylene autoclaving tray, 5 L.
More Details
Cole-Parmer Fluorescent/Incandescent Combination Task Lights
Buy Cole-Parmer Fluorescent/Incandescent Combination Task Lights Items Including:
Fluorescent/Incandescent Combination Task Light, Clamp Mount (YO-41340-40).
More Details
Cole-Parmer Dry Block Temperature Calibrators
Buy Cole-Parmer Dry Block Temperature Calibrators Items Including:
Dry block 230V 700X Series
Dry block 120V 700X Series
Insert Block probe 3/8”
Insert Block probe 5/16”
Insert Block probe 1/4”
More Details
Lab Companion Environmental Chambers
Buy Lab Companion Environmental Chambers Items Including:
Temperature Chamber 20 L, (-20 C To 120°C) 20L; 220V 60 HZ
Temperature Chamber 100 L, (-20 C To 120°C) 100L; 220V 60 HZ
Temperature Chamber 100 L, (-20 C To 120°C) 100L; 230V 50 HZ
Temperature And Humidity Chamber 20 L, (0 C To 120°C) 20L; 220V 60.
More Details
Bel-Art General Accessories and Replacement Items
Buy Bel-Art General Accessories and Replacement Items Items Including:
Replacement filter element for 79774-10 (YO-79774-11)
Slide Storage System, Wheel Set for Five-Drawer Unit (YO-48580-51)
compact flash card reader, USB cable and companion software (YO-80813-75)
128 Megabyte Compact Flash Memory Card (YO-80816-18).
More Details
Ika Lab Equipment
Buy Ika Lab Equipment Items Including:
IKA Lab Dancer Compact Shaker, 100-240V
IKA Continuous feed grinding drive, 230 VAC/50 Hz
Dispersing Element 10mm X 100mm For IKA T-10 Basic Homogenizer (3370100)
Plastic Dispersing Element 10mm X 150mm (10/pk) For IKA T-25 Digital H (YO-04720-91).
More Details
6 Port Manifold
Buy 6 Port Manifold Items Including:
Acetal manifold, 6 port 1/4" NPT(F) inlet, 1/8" NPT(F) outlet (YO-31524-12)
Acetal manifold, 6 port 3/8" NPT(F) inlet, 1/4" NPT(F) outlet (YO-31524-14).
More Details
70618
results
<<<Previous
17
18
19
20
21
22
23
24
25
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
40
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Goggles Safety Uvex to 6 Port Manifold
>
31125329-s-toc_2 31125329stoc2