Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 23

Industrial Water Pumps   to   Spot Beam

Image
  Product Detail
More Detail

Industrial Water Pumps

Buy Industrial Water Pumps Items Including:
  • Pump, Centrifugal Pump, Hot water recirculation pump, 6 GPM, 3/4" Swea
  • Pump, Centrifugal Pump, Hot water recirculation pump, 10 GPM, 3/4" Swe
  • Global Water SP200 Variable Speed, Peristaltic, Fluid Sampling Pump (SP200).

Safety Test Equipment

Buy Safety Test Equipment Items Including:
  • AccTech Cleaning Pads, TexStat® (CT814)
  • Cole-Parmer® Micro-90® cleaning solution, case of twelve 1 qt (YO-18100-00)
  • Cole-Parmer® Micro-90® cleaning solution, 1 quart bottle (YO-18100-01).

Motors Geared

Buy Motors Geared Items Including:
  • Gear Drive System providing turndowns for 1800 to 3600 rpm motors, 0.1
  • Replacement motor for drive system 75211-20
  • Bronze Gear Pump Head Motor Adapter for IEC motors
  • Micropump Cavity-Style Gear Pump Head for NEMA Type 56 C-face Motors, (L20563).

Cole-Parmer Rotors

Buy Cole-Parmer Rotors Items Including:
  • Microcentrifuge Rotor; 24 position, 1.5/2.0mL tubes; for use with 1740
  • Cole-Parmer VS-13000 Microcentrifuge with 6-place 12-mL Tube Rotor, 12
  • Cole-Parmer VS-13000 Microcentrifuge with 6-place 12-mL tube rotor & 2 (C884).

Disposable Needles

Buy Disposable Needles Items Including:
  • BD Bulk Stainless Steel Needles, Non-sterile Clean With Shields, 25g B (YO-07945-62)
  • Portable Disposable Tube Viscometer (PDV-100)
  • Portable Automatic Disposable Tube Viscometer (PDVA-100)
  • B-D™ Needles, 1"L, 0.51 mm OD (YO-07940-52).

100 Ml Burette

Buy 100 Ml Burette Items Including:
  • COLE PARMER BURETTE, 100ML WITH GLASS STOPCOCK
  • COLE PARMER BURETTE, 100ML WITH GLASS STOPCOCK, 4/CASE.

6 Outlets

Buy 6 Outlets Items Including:
  • Standard 6-outlet strip, 6-ft cord (UL207BC)
  • Tripp Lite Three-Stage Surge and Noise Suppressors, 6 outlets
  • PTFE Manifold; 6 Outlets; 1/4" Inlet NPT(F) (YO-31521-13)
  • 304 Stainless Steel manifold; 6 outlets; 1/4" inlet NPT(F) (YO-31521-43).

Oxygen Detectors

Buy Oxygen Detectors Items Including:
  • GasAlertClip™ Extreme Personal Monitor, Oxygen, 2-year operational
  • GasAlert Extreme Single-Gas Detector, Oxygen, with data logging
  • Bacharach GDC-350 Fixed-Mount Gas Monitor, Oxygen
  • Bacharach GDC-350 Fixed-Mount Gas Monitor, Oxygen/Nitrogen dioxide.

Cole-Parmer Zinc

Buy Cole-Parmer Zinc Items Including:
  • Electro-optic point-level sensor; zinc/nickel-plated carbon steel, wet (153843)
  • Electro-optic point-level sensor; zinc/nickel-plated carbon steel, dry (154178)
  • ZINC NITRATE HEXAHYDRATE 9 5G (YO-88226-53)
  • ZINC ACETATE DIHYDRATE R 500G (YO-88289-38).

Gloves for Cleaning

Buy Gloves for Cleaning Items Including:
  • Microflex Latex Cleanroom Glove, ISO Class 4 compliant, extra large, 1
  • Microflex Nitrile Cleanroom Glove, ISO Class 4 compliant, small, 100/b
  • Ansell AccuTech Sterile Latex Gloves, Size 7, 20/BX
  • Ansell AccuTech Sterile Latex Gloves, Size 7.5, 20/BX.

Portable Surface Roughness

Buy Portable Surface Roughness Items Including:
  • Phase-II SRG-4500 Portable Surface Roughness Tester.

One Ultra

Buy One Ultra Items Including:
  • Microflex Ultra One Latex Gloves; size, small
  • Microflex Ultra One Latex Gloves; size, medium
  • Microflex Ultra One Latex Gloves; size, large.

Extraction Spe

Buy Extraction Spe Items Including:
  • Thermo Scientific HyperSep Retain PEP SPE Column, 1mL volume, 30mg bed (YO-01857-06)
  • Thermo Scientific HyperSep C18 SPE Column - 1mL volume, 50mg bed weigh (YO-01857-24)
  • Silica SPE cartridges, 3 mL (YO-10937-00).

Glass Model

Buy Glass Model Items Including:
  • Sintered Glass Base, Kg13 (311002)
  • Sintered Glass Base, Kg25 (311202)
  • Glass Extension 2", Kg47 (311450)
  • Replacement notched glass plate 3/16" thick (YO-28554-54)
  • Notched Glass Plate for Standard Double-Sided Vertical Cells (YO-28554-55).

Gilmont Variable Area Technologies / Rotameters

Buy Gilmont Variable Area Technologies / Rotameters Items Including:
  • Gilmont Variable Area Shielded Flowmeters, sz 10
  • Gilmont Variable Area Shielded Flowmeters, sz 11
  • Gilmont Variable Area Shielded Flowmeters, sz 12
  • Gilmont Variable Area Shielded Flowmeters, sz 13
  • Gilmont Variable Area Shielded Flowmeters, sz 14.

Kevlar Safety Gloves

Buy Kevlar Safety Gloves Items Including:
  • Sperian® Tuff Coat™ Nitrile-Coated Kevlar® Glove, Medium
  • Sperian® Tuff Coat™ Nitrile-Coated Kevlar® Glove, X-Large
  • Sperian® Tuff-Knit KV™ Cut-Resistant Kevlar® Glove, Light (KV13A)
  • Sperian® Tuff-Knit KV™ Cut-Resistant Kevlar® Glove, Light (KV13AL).

Bevco Polyurethane Stools

Buy Bevco Polyurethane Stools Items Including:
  • Clean Room Polyurethane Chair, 16" - 21" Height
  • Polyurethane chair, black, seat height 16" - 21", black reinforced pla (7000-BLACK)
  • Polyurethane chair, black, seat height 22" - 32", black reinforced pla (7500-BLACK).

Strain Gauge Meter

Buy Strain Gauge Meter Items Including:
  • PAXS0000 DIN Strain Gauge / Load Cell Panel Meter 24mV Diff erential 2.

322 Fluke

Buy 322 Fluke Items Including:
  • Fluke 117 Electrician's Combo Kit with 322 Clamp Meter and 115 durable
  • Fluke 62/322/1AC IR Thermometer, Clamp Meter, and Voltage Detector Kit (FLUKE-62/322/1AC)
  • Fluke 322 Clamp Meter (FLUKE-322).

Reversible Temperature Indicating Labels

Buy Reversible Temperature Indicating Labels Items Including:
  • HOT Hand Reversible Indicating Label, Changes from orange to yellow at (4071COPADP10).

Contact Thermometer

Buy Contact Thermometer Items Including:
  • IKA ETS-D6 contact thermometer with pH (3378100)
  • Oakton® Infrared Thermometer with Type-K Input (High Temp) (YO-39644-10).

Chemical Hazard Signs

Buy Chemical Hazard Signs Items Including:
  • NFPA Notice Chemical Hazard, Sign Only, 14" X 10", adhesive vinyl
  • NFPA Notice Chemical Hazard, Sign Only, 14" X 10", aluminum
  • NFPA Notice Chemical Hazard, Sign Kit, 14" X 10", adhesive vinyl.

Chemfluor Fittings

Buy Chemfluor Fittings Items Including:
  • Pipe fittings, Straight coupling, Chemfluor® PTFE, 1/8" NPT(F), 15 (YO-06469-16)
  • Pipe fittings, Reducing bushing, Chemfluor® PTFE, 1/4" NPT(M) x (YO-06469-00)
  • Convoluted FEP-lined hose assembly, 1" hex male, 1-ft length (YO-96550-04).

Ptfe Bottle

Buy Ptfe Bottle Items Including:
  • Cole-Parmer PTFE bottle, 100mL, 1/ea (015.100.2)
  • Cole-Parmer PTFE bottle, 150mL, 1/ea (015.150.2)
  • Cole-Parmer PTFE bottle, 250mL, 1/ea (015.250.2)
  • Cole-Parmer PTFE bottle, 500mL, 1/ea (015.500.2)
  • Cole-Parmer PTFE bottle, 1000mL, 1/ea (015.1000.2).

U V Replacement Bulb

Buy U V Replacement Bulb Items Including:
  • Replacement bulb, 13-watt, for wide-view illuminated magnifiers
  • Replacement Bulb guard for 12-mm electrodes
  • Pack of 4 guards.
  • Replacement bulb for Fiber Optic Illiminator System; 150W; 2000-hour s (YO-09741-02)
  • Replacement bulb for 41200-70 (WTL).

Air Conditioner Accessories

Buy Air Conditioner Accessories Items Including:
  • I/q Rail AC Pwr Dist Asy 2mod
  • I/q Rail DC Pwr Dist Asy 4mod
  • I/q Rail AC Pwr Dist Asy 4mod.

Hp 3 in 1

Buy Hp 3 in 1 Items Including:
  • General Purpose Three-phase TEFC/ODP NEMA Type C-face Motor, 1/3 Hp, 3
  • Lightnin Nettco S-Series Sanitary Mixer, gear-drive, 1-hp, 3-phase, 20 (YO-50323-76)
  • Lightnin Nettco S-Series Sanitary Mixer, gear-drive, 1-hp, 3-phase, 20 (YO-50323-84).

Wave Uv

Buy Wave Uv Items Including:
  • Short/long-wave UV lamp; 4 watts, 254/365 nm wavelength, 115 VAC/60 Hz (95-0021-12)
  • Short/long-wave UV lamp; 4 watts, 254/365 nm wavelength, 230 VAC/50 Hz (95-0021-10)
  • Replacement bulb with 7° beam width for High Intensity UV Lamps (YO-97600-19).

Fluid Filters

Buy Fluid Filters Items Including:
  • Filter top cap for 5 L sealed tubs, 250 mesh (YO-02658-66)
  • Filter top cap for 5-L sealed tub, 500 mesh (YO-02658-67)
  • Filter top cap for 5-L sealed tub, 3 µm/polyester (YO-02658-69)
  • Filter bag for non-sealed tubs, Large, polypropylene (YO-02658-83).

Spot Beam

Buy Spot Beam Items Including:
  • Replacement bulb, spot beam, 15° spread (240-260).



Industrial Water Pumps to Spot Beam

>