Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 25

Cole-Parmer Light-Duty Mixer   to   Portable Temperature

Image
  Product Detail
More Detail

Cole-Parmer Light-Duty Mixer

Buy Cole-Parmer Light-Duty Mixer Items Including:
  • Constant-torque, light-duty mixer, 115 VAC (YO-50402-00)
  • Light-Duty, Dual-Shaft Variable Speed Mixer, 120 VAC (YO-04330-00)
  • Talboys low-cost laboratory mixer; light-duty, variable speed, single (YO-04335-00).

Connector Gasket

Buy Connector Gasket Items Including:
  • SterilEnz sanitary connector, polypropylene with silicone gasket, 1/4" (2AT-408-TC-10)
  • SterilEnz sanitary connector, polypropylene with silicone gasket, 3/8" (2AT-608-TC-10)
  • Mini-gasket for SterilEnz, silicone, pack of 10 (2AT-075).

Hi Torque Motors

Buy Hi Torque Motors Items Including:
  • Test stand, torque, motorized, 100 lbin, vertical, 110V
  • Single Column Motorized Test Stand, 115 VAC
  • Single Column Motorized Test Stand, 230 VAC
  • Motorized, Programmable Test Stand
  • Motorized Force Gauge Test Stand, Horizontal standard (YO-93950-94).

Oakton Economical Probes

Buy Oakton Economical Probes Items Including:
  • Low-Cost RTD PTFE-coated probe; 4.5"L (111897C)
  • Low-Cost RTD surface probe with ceramic tip; 1.4"L (121097A)
  • Low-Cost air/gas RTD probe; 4.5"L (121097B)
  • Low-Cost RTD Probe; General-Purpose, 4.5"L, with ANSI Connector (93831-00).

Cole-Parmer Temperature Labels and Crayons

Buy Cole-Parmer Temperature Labels and Crayons Items Including:
  • Temperature Labels, Refrigerator (YO-90308-80)
  • Temperature Labels, Freezer (YO-90308-85)
  • One-Temperature point indicating labels; temperature point 140°F ( (YO-90309-05)
  • Descending Irreversible Low-Temperature Indicators, 32°F (0°C) (YO-90309-60).

Raytek 1/8-Din Meters

Buy Raytek 1/8-Din Meters Items Including:
  • Standard infrared meter with two 5 VDC alarm outputs (RAYGPC).

E-a-R Personal Protective Equipment (Ppe)

Buy E-a-R Personal Protective Equipment (Ppe) Items Including:
  • Encon Veratti 1000 Protective Eyewear, Clear Lens, Silver Frame
  • 3M personal air monitor, organic vapors, including analysis, 5/bx
  • 3M Personal Air Monitor, Ethylene Oxide, including analysis, 5/bx
  • Ansell TNT® Blue Nitrile Gloves, Powdered, Small.

Hazardous Location

Buy Hazardous Location Items Including:
  • Explosion-proof switch for Hazard Location Motor Base; item numbers 04 (BHLSW)
  • Stainless steel container, 1 liter for Hazard Location Motor Base; ite (HL515).

Super-Nuova Digital Hot Plates

Buy Super-Nuova Digital Hot Plates Items Including:
  • Thermo Scientific Super-Nuova™ aluminum hot plate, 7 x 7" 120vac
  • Thermo Scientific Super-Nuova™ aluminum hot plate, 7 x 7" 220-240v
  • Thermo Scientific Super-Nuova™ ceramic hot plate, 10 x 10" 120VAC.

3000 Psi

Buy 3000 Psi Items Including:
  • 0 to 3000 psi and 0 to 200 kg/cm2 Dual-Scale Gauge, Bottom
  • 0 to 3000 psi and 0 to 200 kg/cm2 Dual-Scale Gauge, Back Co
  • Ashcroft Commercial Gauge with Back Connection, 0 to 3000 psi, 2-1/2"
  • Ashcroft Duragauge Plus! Process Gauge, 0 to 3000 psi.

Thermo Instruments

Buy Thermo Instruments Items Including:
  • Thermo Scientific AquaSensors DataStick AquaClear On-Line Turbidimeter
  • Thermo Scientific AquaPro Multiparameter Analyzer: with Optical RDO Mo
  • Sample position illuminator upgrade for Thermo Scientific NanoDrop ND8 (ND-8K-ILLUM-UP).

Vacuum Solenoid Valves

Buy Vacuum Solenoid Valves Items Including:
  • Vacuum solenoid valve switch (S070C-6CC0-32)
  • Solenoid vacuum valve; right-angle, NW16, aluminum, 110 VAC
  • Solenoid vacuum valve; in-line, NW16, aluminum, 110 VAC
  • Solenoid Vacuum Valve Acid Proof (31F04027)
  • Solenoid Vacuum Valve Chemical (31F04073).

Flir Temperature, Humidity and Moisture Products

Buy Flir Temperature, Humidity and Moisture Products Items Including:
  • Flir B50 Thermal Imaging Infrared Camera With Laser For Building Indus (FLIR B50)
  • Flir B60 Thermal Imaging Infrared Camera With Laser And Scalable Pip F (FLIR B60)
  • Flir T250 Thermal Imaging Infrared Camera With Voice Annotation (76,80 (T250).

Fiber Lights

Buy Fiber Lights Items Including:
  • Cole-Parmer LED Fiber Optic Illuminator, 90 to 264 VAC, 50/60 Hz, 115
  • Cole-Parmer LED Fiber Optic Illuminator with single gooseneck light pi (YO-41723-52)
  • Cole-Parmer LED Fiber Optic Illuminator with single gooseneck light pi (YO-41723-53).

Cole-Parmer Gauss Meters

Buy Cole-Parmer Gauss Meters Items Including:
  • MG-4D-HP-14S : Gaussmeter Package with HP-14S Transverse Probe, Case, (60-B411756C)
  • MG-4D-HP-245S : Gaussmetr Pkg W/Hp245S Axial Probe Case 6FT.Probe Cabl (60-B411756H)
  • Probe Replacement Transverse (121704)
  • Replacement Probe Transverse (121804).

Cole-Parmer Brass Frame/Brass Wire

Buy Cole-Parmer Brass Frame/Brass Wire Items Including:
  • Cole-Parmer Testing Sieve, Brass Frame/SS Wire, 8" OD, full height, No (YO-59940-01)
  • Cole-Parmer Testing Sieve, Brass Frame/SS Wire, 8" OD, half height, No (YO-59941-13).

Large Funnels

Buy Large Funnels Items Including:
  • Large industrial-size low-density polyethylene funnel, 1.5 L (YO-06121-10)
  • Large industrial-size polypropylene funnel, 12 L (YO-06121-00)
  • Large industrial-size polypropylene funnel, 4.5 L (YO-06121-32)
  • Weighing Funnels, Glass, Large (YO-34700-06).

Optical Sensors

Buy Optical Sensors Items Including:
  • Optical Sensor Controller; Allows For Single-point Detection And Contr (149535)
  • Electro-optic point-level sensor; PSF, wet sink, 1/2" straight thread (143580)
  • Electro-optic point-level sensor; PSF, dry sink, 1/2" straight thread (143575).

Msds Compliance

Buy Msds Compliance Items Including:
  • Replacement MSDS Binder, 1 1/2", English (2023)
  • Replacement MSDS Binder, 1-1/2", English/Spanish.

Biohazard Disposal

Buy Biohazard Disposal Items Including:
  • Biohazard Bag 14" x 19", 200 per Case (YO-06494-00)
  • Biohazard Bag 19" x 23", 200 per Case (YO-06494-02)
  • Biohazard Bag 25" x 35", 200 per Case (YO-06494-04)
  • Biohazard Bag 31" x 38", 200 per Case (YO-06494-06).

Hand Sanitizer Dispensers

Buy Hand Sanitizer Dispensers Items Including:
  • Stainless steel foot-activated mobile dispenser.

Pressure Logging

Buy Pressure Logging Items Including:
  • Extech Pressure Meter with SD Card Datalogging in Excel format
  • Pressure Meter with Formatted Data Logging SD Card
  • Fluke 700G/TRACK Data Logging Software and Cable
  • Digital Pressure Gauge with Data Logging and Bar Graph Display, Vacuum (YO-68086-18).

Masterflex Norprene

Buy Masterflex Norprene Items Including:
  • Masterflex Norprene Food A60F Tubing, B/T 87, 25' (YO-06402-87)
  • Masterflex Norprene Food A60F Tubing, B/T 91, 25' (YO-06402-91)
  • Masterflex Norprene Food A60F Tubing, B/T 87, 10 FT (3.0 M)
  • Masterflex Norprene high-pressure tubing, L/S 15HP, 25 ft..

Whatman Syringe Filters

Buy Whatman Syringe Filters Items Including:
  • Whatman Anotop syringe filter, 10mm, 0.1µm, sterile, 50/pk (6809-1112)
  • Whatman 25-mm GD/X Syringe Filters, Surfactant-free cellulose acetate; (YO-29705-00)
  • Whatman 25-mm GD/X Syringe Filters, Surfactant-free cellulose acetate; (YO-29705-04).

Jars Plastic

Buy Jars Plastic Items Including:
  • Thermo Scientific Nalgene multipurpose polycarbonate jar with lid, 2 1
  • Thermo Scientific Nalgene multipurpose polycarbonate jar with lid, 1 1
  • Thermo Scientific Nalgene multipurpose polycarbonate jar with lid, 2 q.

Nor-Lake Safety Refrigerators/Freezers

Buy Nor-Lake Safety Refrigerators/Freezers Items Including:
  • Refrigerator, Flammable Storage 24 cu ft with Casters, 115V/60Hz (NSFR241WMW/0M).

Soil Hydrometers

Buy Soil Hydrometers Items Including:
  • H-B Instrument Soil Hydrometer, ASTM 151H, 0.995 to 1.038 Specific Gra.

Lufkin Measuring Tapes

Buy Lufkin Measuring Tapes Items Including:
  • Lufkin 1" x 25' Hi-Viz Orange Power Tape (YO-97105-86)
  • Lufkin 3/4" x 12' Hi-Viz Orange Series 1000 Power Tape (YO-97105-88).

Cole-Parmer Multifunction Dataloggers

Buy Cole-Parmer Multifunction Dataloggers Items Including:
  • Dual-channel multifunction datalogger
  • Universal Input Datalogger (ES120)
  • Simple Logger II Model L404 (4-Channel Event)
  • Pressure logger with display, 0 to 300 PSI
  • Pressure logger with display, 0 to 500 PSI

Portable Temperature

Buy Portable Temperature Items Including:
  • Portable heat sealer, 220 VAC, 50/60 Hz (1F02200)
  • Shuttle Portable Ultra-Low Temperature Freezer, 120/220V US std
  • Plug
  • DAQ-PRO - 8 Channel Logger: Voltage/Current/Temperature
  • Includes Soft



Cole-Parmer Light-Duty Mixer to Portable Temperature

>