Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 26
Cast Iron Pump
to
Chrome Wire Shelving
Image
Product Detail
More Detail
Cast Iron Pump
Buy Cast Iron Pump Items Including:
Flanges for 79730-00, 79730-10, 79731-00 pump, cast iron 1" size
Flanges for 79730-00 pump, cast iron 1-1/4" size
Flanges for 79732-00 and -04 pumps, cast iron 3/4" size
Flanges for 79732-00 and -04 pumps, cast iron 1-1/4" size.
More Details
Liquid Samplers
Buy Liquid Samplers Items Including:
Drum and Liquid Sampler, 3', HDPE (114505)
Drum and Liquid Sampler, 6', HDPE (114515-0500)
Drum and Liquid Sampler, 8', HDPE (114525)
Dipstik™ PTFE Drum And Tank Sampler, 3.5'long (D8653242)
Dipstik™ PTFE Drum And Tank Sampler, 7' Long (D8653274).
More Details
Scienceware Polyfoil Square Wrap
Buy Scienceware Polyfoil Square Wrap Items Including:
Polyfoil Square Wrap 4" X 4" (YO-06275-02)
Polyfoil Square Wrap 5 1/2" X 5 1/2" (YO-06275-04).
More Details
Autovial Syringe Filters
Buy Autovial Syringe Filters Items Including:
Whatman Autovial Syringeless Filters, Nylon; 0.20µ; 50/box
Whatman Autovial Syringeless Filters, Nylon; 0.45µ; 50/box
Whatman Autovial Syringeless Filters, PTFE; 0.20µ; 50/box
Whatman Autovial Syringeless Filters, PTFE; 0.45µ; 50/box.
More Details
H-B Instruments
Buy H-B Instruments Items Including:
Eutech Instruments pH 200 1/8-DIN pH/ORP controller
Eutech Instruments pH 1000 controller, 115 VAC
Eutech Instruments pH 1000 controller, 220 VAC
Eutech Instruments pH 2000D, differential pH/ORP Controller
Eutech Instruments pH 550 pH/orp Monitor.
More Details
Industrial Instrumentation
Buy Industrial Instrumentation Items Including:
Instrument Cabinet, 40" high
(30-2301-GT)
Cabinet, for 19" wide Instruments (ER-16525)
Bud Industries TR-Series Enclosure for DIN-Mount Controllers, Small Si
Cole-parmer Industrial Pressure Gauge, 30" Hg To 30 PSI; 1/4" NPT(M).
More Details
Disposable Weighing Dishes
Buy Disposable Weighing Dishes Items Including:
Aluminum weighing dishes with tab, 43 mm (80064)
Aluminum weighing dishes with tab, 57 mm (80065)
Aluminum weighing dishes with tab, 70 mm (80065L)
Aluminum weighing dishes, 70 mm, Smooth (80065S)
Aluminum Dishes, Disposable
More Details
Cole-Parmer Housings for 20
Buy Cole-Parmer Housings for 20 Items Including:
Cole-Parmer General Purpose pH Electrode, 10-ft Cable
Cole-Parmer self-cleaning pH electrode, Ryton housing, 10-ft cable, BN
Cole-Parmer Removable High pH Double-Junction pH Electrode
In-Line pH Electrodes with CPVC Housing, 1 kΩ RTD (YO-27106-20).
More Details
Colored Caps
Buy Colored Caps Items Including:
Cyrogenic vial; 2.0 mL, external thread, flat bottom, blue-colored cap
Cyrogenic vial; 2.0 mL, external thread, flat bottom, pink-colored cap
Cyrogenic vial; 2.0 mL, external thread, flat bottom, red-colored cap.
More Details
Tyvek Safety Apparel
Buy Tyvek Safety Apparel Items Including:
Tyvek Boot Cover, 100 pair/case
Tyvek Coveralls, Large
Tyvek Coveralls, X-Large
Tyvek Coveralls, XX-Large
Tyvek Coveralls with Elastic Cuffs, Medium
More Details
Histology Preparation
Buy Histology Preparation Items Including:
Slide warmer, 115 VAC 50/60 Hz (YO-48573-01)
Slide warmer, 230VAC 50/60Hz
100-ml Mounting Media Bottle (YO-48585-62)
Staining dish and cover (YO-48585-70)
Stainless steel rack for 30 slides (YO-48585-76)
More Details
Colder Multi-Tube Connectors
Buy Colder Multi-Tube Connectors Items Including:
Tubing Shroud for Ten-Line Tubing Connector (TS10)
Coupling Insert, Two-Line, Acetal, 1/16" ID Tube (PTC22010)
Coupling Insert, Two-Line, Acetal, 1/8" ID Tube (PTC22020)
Coupling Body, Six-Line, Male, Acetal, 1/16" ID Tube (SXM1701).
More Details
Boekel Incubator
Buy Boekel Incubator Items Including:
Boekel Jitterbug Microplate Incubator Shaker, 115 VAC, 50/60 Hz (YO-13059-00).
More Details
Multimeter Tester
Buy Multimeter Tester Items Including:
Fluke-179/1AC-II Multimeter, Voltage Tester & Acc
Kit (FLUKE-179/1AC-II)
Dranetz Insulation Tester/TRMS Digital Multimeter/Datalogger
Fluke-62 MAX+/T+PRO/1AC Electrical/HVAC Combo Kit
Fluke T5-600/62MAX+/1AC Electrical/IR Kit.
More Details
Metal Detectors
Buy Metal Detectors Items Including:
Thermo Scientific APEX 100 Metal Detector, with 350 mm width x 350 mm
Thermo Scientific APEX 100 Metal Detector, with 350 mm width x 150 mm
Thermo Scientific APEX 100 Metal Detector, with 500 mm width x 200 mm.
More Details
Cole-Parmer Ammonium Acetate
Buy Cole-Parmer Ammonium Acetate Items Including:
ACETIC ACID AMMONIUM SALT 25G (YO-88360-56).
More Details
I/P Food Quality & Testing
Buy I/P Food Quality & Testing Items Including:
Masterflex® L/S® variable-speed drive with 10-turn speed contr (77521-57)
Costar small scraper, 18 mm blade x 250 mm handle
MiniStrip film, 200/pk (YO-13024-04)
MiniStrip film, 200/pk, sterile (YO-13024-06)
AccTech Cleaning Pads, TexStat® (CT814).
More Details
Proximity Sensing
Buy Proximity Sensing Items Including:
Inductive, M8, 1.5mm sensing distance, NPN-NO, shielded
Inductive, M12, 2mm sensing distance, NPN-NO, shielded
Inductive, M18, 5mm sensing distance, NPN-NO, shielded (E2E-X5E1)
Inductive, M12, 2mm sensing distance, NPN-NO, shielded proximity senso.
More Details
Fluid Handling
Buy Fluid Handling Items Including:
Microfluidic Fluid Splitter Slide, Several splitters on one slide
Microfluidic Micro Fluid Connector, COC material, package of 25 pieces
Flexible shaft for overhead stirrers (YO-54400-45)
Splash protector for Heidolph overhead mixers (YO-54400-50).
More Details
Fume Extraction Hoods
Buy Fume Extraction Hoods Items Including:
Extract-All Portable Air Cleaning System with 4' to 6' Long Extractor
Secondary Merv 8 Pleated Filter
Primary Merv 14 Cell Filter.
More Details
Cole-Parmer Hazardous Materials Training Posters
Buy Cole-Parmer Hazardous Materials Training Posters Items Including:
Right-to-Know training poster, 24" X 28".
More Details
Gas Detection Instruments
Buy Gas Detection Instruments Items Including:
VOC-TRAQ™ USB PID Toxic Gas Detector, 1.0 to 1000 ppm (isobutylene)
Multi Pro 3 Gas Detector, O2 LEL CO
Multi Pro 4 Gas Detector, O2 LEL Duo-Tox CO/H2S
GasAlertMaxXT™ Four Gas Detector
Rae Four-Gas Personal Protection Monitor, Datalogging Monitor.
More Details
Count Up Timers
Buy Count Up Timers Items Including:
Cole-Parmer Walkaway™ Count-Up Controller, 0.01 sec to 99 hrs
Cole-Parmer Walkaway™ Count-Up Controller, 1 sec to 99 hrs.
More Details
Group Lockout Boxes
Buy Group Lockout Boxes Items Including:
Group View Slot Lockout Box, Yellow (KCC630).
More Details
Thermo Scientific Storage Equipment
Buy Thermo Scientific Storage Equipment Items Including:
Floorstand for Thermo Scientific Barnstead Metal Classic Stills Storag (H1000)
Thermo Scientific Nalgene Standard Cryocane, Aluminum, 300 mL (holds 6
Thermo Scientific Nunc™ CryoTube Vial Holder
Thermo Scientific Nunc Chipboard Storage Box, 6/case.
More Details
Parker Hannifin Female Threaded Adapters (Straight)
Buy Parker Hannifin Female Threaded Adapters (Straight) Items Including:
Compression fittings, female pipe adapter, straight, Black PP, 1/4" OD (YO-06444-11)
Compression fittings, female pipe adapter, straight, Black PP, 3/8" OD (YO-06444-12).
More Details
Room Monitors
Buy Room Monitors Items Including:
Room Pressure Monitor/Alarm, ±0.25" WC, 24 VAC
Barometric pressure monitor, 600 to 1100 hPa/mb
Absolute pressure monitor, 0 to 20 psia
Room Pressure Monitor/Alarm, ±0.5" WC, 24 VAC
Room Pressure Monitor/Alarm, ±0.5" WC, 120/240 VAC.
More Details
Pocket Loupe
Buy Pocket Loupe Items Including:
Bausch & Lomb Folding Pocket Magnifier, single lens, 4x magnification
Bausch & Lomb Folding Pocket Magnifier, dual lens, 4x to 9x magnificat.
More Details
Cole-Parmer Portable Benchtop Chilling Incubator
Buy Cole-Parmer Portable Benchtop Chilling Incubator Items Including:
Portable Benchtop Chilling Incubator, 100/240VAC (YO-39064-21).
More Details
Chrome Wire Shelving
Buy Chrome Wire Shelving Items Including:
Wire Shelving,18w X 36l X 64h (MA1836C64S-4)
Wire Shelving,18w X 42L X 64h, 4 Shelves (MA1842C64S-4)
Shelf Add-on,18w X 48l X 64h (MA1848C64A-4)
Shelf Add-on,18w X 60l X 64h (MA1860C64A-4)
Shelf Add-on,18w X 72L X 64h (MA1872C64A-4).
More Details
70618
results
<<<Previous
22
23
24
25
26
27
28
29
30
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
36
37
38
39
40
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Cast Iron Pump to Chrome Wire Shelving
>
31125329-s-toc_2 31125329stoc2