Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 27
Digital Oscilloscopes
to
5 Gallon Pail Heater
Image
Product Detail
More Detail
Digital Oscilloscopes
Buy Digital Oscilloscopes Items Including:
Tektronix TDS1002C-EDU Digital Oscilloscope, 2 Channel, 60 MHz, 1 GS/s
Tektronix TDS1012C-EDU Digital Oscilloscope, 2 Channel, 100 MHz, 1 GS/
B&K Precision 2530B Digital Oscilloscope, 25 MHz
B&K Precision 2532B Digital Oscilloscope, 40 MHz.
More Details
Cole-Parmer Batteries
Buy Cole-Parmer Batteries Items Including:
Cole-Parmer Battery-powered Digital Gauge, 0 to 30" Hg; 1/4" NPT(M)
Cole-Parmer Battery-powered Digital Gauge, -30" Hg to 15.0 PSI; 1/4" N
Cole-Parmer Battery-powered Digital Gauge, 0 to 15.00 PSI; 1/4" NPT(M).
More Details
Salmonella Test
Buy Salmonella Test Items Including:
Salmonella test chip, 25 tests/pk (YO-36152-70)
Rapid test for Salmonella (YO-36151-00)
Salmonella enrichment broth according to Rappaport-Vassiliadis (YO-36152-74)
RAMBACH agar, isolation media, 4 x 1L (YO-36152-76).
More Details
Lightnin Axial Flow Impellers
Buy Lightnin Axial Flow Impellers Items Including:
Lightnin® 316 SS axial flow impeller (Lightnin® A-100), 3.6" d (YO-04317-90).
More Details
Flow Control Fluid
Buy Flow Control Fluid Items Including:
Pump, Metering with 5 control methods, 0.66gph, PVDF, Fkm Front Contro (96470133).
More Details
Bel-Art Bottles
Buy Bel-Art Bottles Items Including:
Scienceware low-density polyethylene wide-mouth sample jar, 180 mL 6/
Scienceware polypropylene mason jar, 480 mL
Scienceware polypropylene mason jar, 960 mL
Scienceware polypropylene mason jar, 2 L
Scienceware polypropylene mason jar, 4 L.
More Details
N-Tron Environmental Testing
Buy N-Tron Environmental Testing Items Including:
Ntps2413 : N-tron Pwer Sup 24-1.3 Amp.
More Details
Timer 220
Buy Timer 220 Items Including:
Cole-Parmer Low-Cost Ultrasonic Cleaner with Timer, 220 VAC (YO-08848-15)
Microplate Orbital Shaker with Timer, 230 VAC, 50 Hz.
More Details
Stainless Steel Pressure Switch
Buy Stainless Steel Pressure Switch Items Including:
Pressure Switch, Stainless Steel, 30"hg To 15 PSI; 1/4" NPT(M)
Pressure Switch, Stainless Steel, 0 To 1000 PSI; 1/4" NPT(M)
Stainless steel liquid flow switch for high inline pressures; 0.25 GPM (YO-32774-40).
More Details
Chemware Plastic Flasks
Buy Chemware Plastic Flasks Items Including:
Chemware Erlenmeyer Flask, PFA, 100 mL
Chemware Erlenmeyer Flask, PFA, 200 mL.
More Details
Load Sensors
Buy Load Sensors Items Including:
Slc 50 X 0.01 Lbf Load Sensor Smart Technology
Alc 2 Lbf X 0.0002 Lbf Load Sensor Smart Technology
Slc 10 Lbf X 0.001 Lbf Load Sensor Smart Technology
Slc 100 X 0.01 Lbf Load Sensor Smart Technology
Slc 200 X 0.02 Lbf Load Sensor Smart Technology.
More Details
Electronic Indicator
Buy Electronic Indicator Items Including:
Electronic depth micrometer (54-225-456)
Electronic depth gauge (54-125-777)
IP65 electronic mini indicator (54-520-680)
2-200 : Analog Wind Speed System.
More Details
Metric Fitting Laboratory Equipment
Buy Metric Fitting Laboratory Equipment Items Including:
304 SS four-blade propeller for shaft 04370-02, 75 mm propeller dia (YO-04370-11)
304 SS four-blade propeller for shaft 04370-02, 60 mm propeller dia (YO-04370-12)
304 SS folding propeller for shaft 04370-02, 48 mm propeller dia (open (YO-04370-14).
More Details
Cole-Parmer High-Flow Nylon Monofilament Filter Bags
Buy Cole-Parmer High-Flow Nylon Monofilament Filter Bags Items Including:
Size 1 high-flow nylon monofilament filter bag; 300°F (149°C) (255050-03)
Size 1 high-flow nylon monofilament filter bag; 300°F (149°C) (255051-03)
Size 1 high-flow nylon monofilament filter bag; 300°F (149°C) (255052-03).
More Details
Extech Economical Digital Tachometers
Buy Extech Economical Digital Tachometers Items Including:
Economical Digital Contact Tachometer (461891)
Economical Digital Optical Tachometer (461893).
More Details
Plate Shakers
Buy Plate Shakers Items Including:
Thermo Scientific Titer Plate Shaker, 120 VAC
Thermo Scientific Titer Plate Shaker, 240 VAC
Double-tier 96-well Microwell Plate Insert for Vortex-Genie 2™ (SI-0510).
More Details
Gillis Lpi Furniture & Fume Hoods
Buy Gillis Lpi Furniture & Fume Hoods Items Including:
Wire Shelving,18w X 36l X 64h (MA1836C64S-4)
Wire Shelving,18w X 42L X 64h, 4 Shelves (MA1842C64S-4)
Wire Shelving,18w X 48l X 72h (MA1848C72S-4)
Wire Shelving,18w X 60l X 72h (MA1860C72S-4)
Wire Shelving,18w X 72L X 72h (MA1872C72S-4).
More Details
Orbeco-Hellige Turbidity Handheld Meters
Buy Orbeco-Hellige Turbidity Handheld Meters Items Including:
Orbeco-Hellige TB200-IR Handheld Turbidity Meter, ISO compliant.
More Details
Replacement Membranes
Buy Replacement Membranes Items Including:
Replacement membrane filter, 3 µm
Pack of 10 (22 23 110-3)
Standard DO membrane kit for DO probe 05521-00, pack of 30 membranes a (5775)
Replacement membrane kit for DO probes
Cole-Parmer Inline Bubble trap, 5 Pack of Replacement Membranes for 75.
More Details
Cole-Parmer Tripod Bases
Buy Cole-Parmer Tripod Bases Items Including:
Tripod Bases, tripod f/03218-50 -52 (YO-03218-58)
Tripod Bases, tripod f/03218-54 -56 (YO-03218-59)
Tripod Base for 3- or 5-tube Models (YO-03226-40)
Tripod Bases, tripod base f/multitube frames (YO-03226-50).
More Details
Amresco Reagents
Buy Amresco Reagents Items Including:
Mounting media, 120-mL bottle (YO-48804-64)
Agarose III™ Electrophoresis Solution, 25g (YO-21850-60).
More Details
I-Chem Jars
Buy I-Chem Jars Items Including:
I-CHEM pre-cleaned sample jars with certificate of analysis, 1250 mL, (341-1250).
More Details
Cole-Parmer Material Storage and Handling
Buy Cole-Parmer Material Storage and Handling Items Including:
Storage bin; 4-1/8" x 3" x 5-3/8", 24/pack (30-210 YELLOW)
Storage bin; 4-1/8" x 3" x 7-3/8", 24/pack (30-220 YELLOW)
Storage bin; 5-1/2" x 5" x 10-7/8", 12/pack (30-230 YELLOW).
More Details
Interscience Laboratories Inc Reciprocating Paddle Blenders
Buy Interscience Laboratories Inc Reciprocating Paddle Blenders Items Including:
Paddle Blender; 5 To 100 mL, Variable Speed, Window Door, Removable Pa (YO-36850-09)
Paddle Blender; 50 To 400 mL, Fixed Speed, Stainless Steel Door (YO-36850-10)
Paddle Blender; 50 To 400 mL, Variable Speed, Window Door, Removable P (YO-36850-13).
More Details
High Intensity Uv
Buy High Intensity Uv Items Including:
Replacement bulb with 7° beam width for High Intensity UV Lamps (YO-97600-19)
Cole-Parmer 25-watt Dual-Intensity 254nm Wavelength UV Transilluminato (YO-97622-00)
Cole-Parmer 25-watt Dual-Intensity 254nm Wavelength UV Transilluminato (YO-97622-02).
More Details
Cole-Parmer Miniature Vials with Hinged Caps
Buy Cole-Parmer Miniature Vials with Hinged Caps Items Including:
Miniature vials with hinged caps, 0.3 mL (EP1539LG-PE)
Miniature vials with hinged caps, 2.1 mL (EP2100LG-PE)
Miniature vials with hinged caps, 16.9 mL, 1000/PK (EP191LG-PE)
Miniature vials with hinged caps, 29.6 mL (EP192LG-PE).
More Details
Glass Insulation
Buy Glass Insulation Items Including:
Thermocouple,Wire,TYPE-K,20-GAUGE, High Temp Glass Braid Insulation, 1 (K20-2-321* (1300F MAX)) (YO-08541-23)
Thermocouple Wire,type-j,24-gauge, Glass Braid Insulation W/SS Overbra (J24-1-S-305 | 08541-35).
More Details
Cole-Parmer Density/Specific Gravity
Buy Cole-Parmer Density/Specific Gravity Items Including:
Digital Density / Specific Gravity Meter
Specific gravity cup with Tare Weight and NIST certificate, 100 mL cap (WG-SS-100/T/C)
SBS-2002 : Digital Hydrometer (SBS-2002)
Pycnometer, 11.5-mL capacity, 1/8" (3.2-mm) hole in lid (03247Q).
More Details
Power Supply Products
Buy Power Supply Products Items Including:
Power supply/charger, 115 VAC for analyzer
Includes four NiCd batter (0554 1085)
9/36 VDC power supply option
Extech Analog DC Power Supply, 115/220 VAC
Quad Output DC Power Supply (382270)
More Details
5 Gallon Pail Heater
Buy 5 Gallon Pail Heater Items Including:
Pail heater, 5-gallon, 200 to 400°F, 1500 watts,120 VAC (TRX-5-H/R)
3" wide flexible drum heater, 5 gallon, 300 watt, 120 VAC, 70-425° (YO-09200-00)
9 1/2" wide flexible drum heater, 5 gallon, 1000 watt, 120 VAC, 70-190 (YO-09203-00).
More Details
70618
results
<<<Previous
23
24
25
26
27
28
29
30
31
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
36
37
38
39
40
41
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Digital Oscilloscopes to 5 Gallon Pail Heater
>
31125329-s-toc_2 31125329stoc2