Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 28
Omni International Inc Omni Mixer Homogenizers
to
Water Sampling Supplies
Image
Product Detail
More Detail
Omni International Inc Omni Mixer Homogenizers
Buy Omni International Inc Omni Mixer Homogenizers Items Including:
Homogenizer Probes, 7mm x 110mm for hard tissue samples (50/box) (YO-04727-44).
More Details
Foam Supplies
Buy Foam Supplies Items Including:
Indenti-plug Plastic Foam Stopper, 6-13mm Openings, 1000/pk
Indenti-plug Plastic Foam Stopper, 21-26mm Openings, 400/pk
Indenti-plug Plastic Foam Stopper, 46-50mm Openings, 80/pk
Foam Laboratory Brush Set
Universal Foam Glassware Brush, 15"L.
More Details
Industrial Equipment
Buy Industrial Equipment Items Including:
Bud Industries Showcase™ Series Enclosure for 1/8 DIN Mount Control (BB-1800)
Bud Industries TR-Series Enclosure for DIN-Mount Controllers, Small Si
Bud Industries TR-Series Enclosure for DIN-Mount Controllers, Medium S.
More Details
Cole-Parmer Auger Kits
Buy Cole-Parmer Auger Kits Items Including:
Soil auger kit (PN077)
Shallow depth backsaver soil sampling kit (PN023)
Medium depth backsaver soil sampling kit (PN019).
More Details
Polypropylene Membranes
Buy Polypropylene Membranes Items Including:
Whatman™ Disposable Capsule Filters, Polypropylene Membrane; pore s (YO-29700-20)
Whatman™ Disposable Capsule Filters, Polypropylene Membrane; pore s (YO-29700-22)
Thermo Scientific Nalgene Disposable Sterile Filter Units; membrane, w (YO-06730-02).
More Details
Blue Cap
Buy Blue Cap Items Including:
Filter cartridge, "Big Blue", 10", carbon block, 0.5 micron (155170-43(CBC-BB))
Filter cartridge, "Big Blue", 20", carbon block, 0.5 micron (155312-43(CBC-20BB))
HART Smart Connector, TC (Blue Cap), Spare (YO-93202-60).
More Details
Nema Explosion Proof
Buy Nema Explosion Proof Items Including:
Explosion-proof Single-phase NEMA Type C-face Motor, 1/4 Hp, 1800 RPM,
Explosion-proof Single-phase NEMA Type C-face Motor, 1/3 Hp, 1800 RPM,
Explosion-proof Single-phase NEMA Type C-face Motor, 1/2 Hp, 3600 RPM,.
More Details
Raytek Temperature Infrared Thermometers / Sensors
Buy Raytek Temperature Infrared Thermometers / Sensors Items Including:
Comapact CM-Series Infrared Sensor Type-J T/C Output with 3' Cable (RAYCMLTJ).
More Details
Cole-Parmer Test Tube Basket
Buy Cole-Parmer Test Tube Basket Items Including:
Test Tube Basket for Cole-Parmer Roto-Torque Variable Speed Rotator.
More Details
Retriever Chromatography Products
Buy Retriever Chromatography Products Items Including:
ISCO Retriever 500 Mini Fraction Collector, 12 to13 mm tubes
ISCO Retriever 500 Mini Fraction Collector, 10 to16 mm tubes
ISCO Retriever 500 Mini Fraction Collector, 18 mm tubes or 17 mm scint.
More Details
Davis Instruments Weather Stations
Buy Davis Instruments Weather Stations Items Including:
Davis Instruments 7654 Solar-powered Long-range Repeater
Davis Instruments 6625 Vantage Pro2 Power Adapter
Davis Instruments 6323 Vantage Pro2 Wireless ISS with 24-hour Fan-Aspi
Davis Instruments 7345.976 Wireless Transmitter Printed Circuit Board.
More Details
Centrifuge Speed
Buy Centrifuge Speed Items Including:
High-speed fixed-angle rotor; six x 85 mL tubes (YO-02750-60)
Fixed-angle Rotors for 17304-series centrifuges (YO-17302-52)
Brushless Variable Speed Ready DC Magnetic Drive Pump, High-Head Polyp
8-place Clinical Centrifuge, variable speed - 220VAC.
More Details
Test Multimeter
Buy Test Multimeter Items Including:
Fluke TLK289 Industrial Meter Test Lead Set (TLK289)
Fluke TLK287 Electronics Master Test Lead Set (TLK287)
Fluke AC283 SureGrip Pin-Grabber Test Clips
Flexible Test Leads
Fluke TL75 Right-Angle Test Lead Set
More Details
Plastic Containers
Buy Plastic Containers Items Including:
Rubbermaid Storage Containers with Handle, White HDPE, 2 quart
Rubbermaid Storage Containers with Handle, White HDPE, 4 quart
Rubbermaid Storage Container with Handles, Clear PP, 2 quart
Rubbermaid Storage Container with Handles, Clear PP, 4 quart.
More Details
Nalgene Labware
Buy Nalgene Labware Items Including:
Thermo Scientific Nalgene high-density polyethylene wide-mouth sample (2189-0001)
Thermo Scientific Nalgene high-density polyethylene wide-mouth sample (2189-0004)
Thermo Scientific Nalgene high-density polyethylene narrow-mouth sampl (2089-0001).
More Details
Autovial Laboratory Filtration Products
Buy Autovial Laboratory Filtration Products Items Including:
Whatman Autovial Syringeless Filters, Nylon; 0.20µ; 50/box
Whatman Autovial Syringeless Filters, Nylon; 0.45µ; 50/box
Whatman Autovial Syringeless Filters, PTFE; 0.20µ; 50/box
Whatman Autovial Syringeless Filters, PTFE; 0.45µ; 50/box.
More Details
Ceramic Furnaces
Buy Ceramic Furnaces Items Including:
Ceramic Ribbed Plate For Furnace Model Lt 3 (691600507)
Ceramic Ribbed Plate For Furnace Model Lt 24 (691600874)
Ceramic hearth plate for 130 cu-in furnaces (9353053)
Ceramic Hearth Plate for 1750 cu in Furnace with Single Set Point Cont (9353060).
More Details
Micronit Microfluidics
Buy Micronit Microfluidics Items Including:
Microfluidic Stainless Steel Connection Kit for Lab-on-chip experiment
Microfluidic Microreactor chip set, 700 micrometer bottom glass with 0 (FC_R150.332.2_PACK)
Microfluidic Micromixer chip set, 700 micrometer bottom glass with 0.2 (FC_SW11_PACK).
More Details
Hydrometer Soil Analysis
Buy Hydrometer Soil Analysis Items Including:
H-B Instrument Soil Hydrometer, ASTM 151H, 0.995 to 1.038 Specific Gra.
More Details
Mini Bead Beater
Buy Mini Bead Beater Items Including:
Mini bead beater, 115 VAC 60 Hz (YO-36270-02)
Mini bead beater, 230 VAC 50 Hz (YO-36270-07)
Glass Beads, 0.1 to 0.15 mm (YO-36270-52)
Zirconia-Silicate Beads, 0.5 mm (YO-36270-61)
100 Sterile Vials with caps in foam rack, 2-mL (YO-36270-70).
More Details
Omni International Inc Homogenizers
Buy Omni International Inc Homogenizers Items Including:
Homogenizer Probes, 7mm x 110mm for hard tissue samples (50/box) (YO-04727-44)
Aerosol-Sealed Assembly with 303 stainless steel mixing chamber; sampl (YO-04729-02)
Aerosol-Sealed Assembly with 303 stainless steel mixing chamber; sampl (YO-04729-06).
More Details
Lmi Metering Pumps
Buy Lmi Metering Pumps Items Including:
LMI Solenoid Metering Pump Service Kit for 74522-*6
LMI Solenoid Metering Pump Service Kit for 74522-*8
Digital Remote-Control Solenoid-Diaphragm Metering Pump, 2.5 GPH, 115
Digital Remote-Control Solenoid-Diaphragm Metering Pump, 0.42 GPH, 115.
More Details
Wall Mount Network Rack
Buy Wall Mount Network Rack Items Including:
3-Part EL Wall-mnt enclosure, 24x24x24 (9969580)
3-Part EL Wall-mnt enclosure, 29x24x24 (9969581)
3-Part EL Wall-mnt enclosure, 40x24x24 (9969582)
DK TS8 Enclosure 79x24x24 42U units (9969880)
DK TS8 Enclosure 79x24x32 42U units (9969892).
More Details
Co2 Electrodes
Buy Co2 Electrodes Items Including:
CO2 Electrode with 12-mm body diameter and 10-ft cable with (YO-29000-01)
CO2 Electrode with 12-mm body diameter and 10-ft cable with (YO-29000-03)
CO2 Electrode with 12-mm body diameter and 10-ft cable with (YO-29000-05).
More Details
Cole-Parmer Dispensers
Buy Cole-Parmer Dispensers Items Including:
Cole-Parmer Console Digital Dispensing Drive with Remote Control for M (YO-75211-30)
Cole-Parmer NEMA 4X Wall-Mount Digital Dispensing Drive, 115 VAC
Cole-Parmer Micro Dispensing Pump, 4-Channel, 115/230 VAC
Cole-Parmer Bottletop Dispenser .5-5 mL.
More Details
Steel Steam
Buy Steel Steam Items Including:
Stainless Steel Steam Generator; 36 Lbs/hr, 208 VAC (SSB12100B3)
Stainless Steel Steam Generator; 36 Lbs/hr, 480 VAC (SSB12100F3)
Stainless Steel Steam Generator; 54 Lbs/hr, 208 VAC (SSB18100B3).
More Details
Pulsafeeder Accessories and Replacement Items
Buy Pulsafeeder Accessories and Replacement Items Items Including:
Service Kit for Solenoid Metering Pump, 6 GPD
Service Kit for Solenoid Metering Pump, 12 GPD
Service Kit for Solenoid Metering Pump, 21- 44 GPD
Service Kit for Solenoid Metering Pump, 94 GPD
Service Kit for Solenoid Metering Pump, 240 GPD.
More Details
High Speed Mixer
Buy High Speed Mixer Items Including:
High-torque, constant-speed mixer, 115 VAC (YO-50402-20)
High-torque, constant-speed mixer, 220 VAC (YO-50402-25)
IKA Eurostar 20 High-Speed Digital Mixer, 150 to 6000 rpm, 115 VAC
IKA Eurostar 20 High-Speed Digital Mixer, 150 to 6000 rpm, 230 VAC.
More Details
Mixed Cellulose Esters
Buy Mixed Cellulose Esters Items Including:
Advantec Mixed Cellulose Ester Membranes; 13 mm dia; 0.20 µm; 100/p
Advantec Mixed Cellulose Ester Membranes; 13 mm dia; 0.45 µm; 100/p
Advantec Mixed Cellulose Ester Membranes; 25 mm dia; 0.20 µm; 100/p.
More Details
Water Sampling Supplies
Buy Water Sampling Supplies Items Including:
Whirl-Pak stand-up sodium thiosulfate bags for potable water sampling, (YO-06499-77)
Whirl-Pak Coli-Test sodium thiosulfate bags for water sampling, 4 oz
Whirl-Pak stand-up sodium thiosulfate bags for potable water sampling, (YO-06499-97).
More Details
70618
results
<<<Previous
24
25
26
27
28
29
30
31
32
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
31
32
33
34
35
36
37
38
39
40
41
42
50
60
70
80
90
100
110
120
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Omni International Inc Omni Mixer Homogenizers to Water Sampling Supplies
>
31125329-s-toc_2 31125329stoc2