Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 3
Ion Filter
to
Cole-Parmer Ptfe Filters
Image
Product Detail
More Detail
Ion Filter
Buy Ion Filter Items Including:
Filter modules for use with Turbidity meters, item numbers 99512-10, - (30397-00)
Mounting bracket kit for Ion-X-Changer Filter Cartridges (YO-01506-60)
Cartridge,adsorber + (D502121)
Cartridge,universal + (D502119)
Cartridge,research + (D502122).
More Details
Handheld 10 Meter
Buy Handheld 10 Meter Items Including:
YSI 30 handheld conductivity meter with cell and 10-ft cable
(030130)
Oakton Waterproof pH/Con 10 Meter with pH/conductivity probe
WTW Multi 3410 Multiparameter Meter w/TetraCon 925-3 probe
Oakton Temp-10 type J thermocouple thermometer with boot.
More Details
Canon Mm
Buy Canon Mm Items Including:
C-mount adapter for Canon 35-mm camera (MA150/50+MA512+T2-1)
C-mount adapter for Canon EOS 35-mm camera (MA150/50+MA512+T2-9)
10:1 Stereozoom Microscope 35-mm Camera Adapter, Canon (T2-1).
More Details
Chemsaver Dispensers
Buy Chemsaver Dispensers Items Including:
Brinkmann ChemSaver Dispenser 0.1-2.5 mL
Brinkmann ChemSaver Dispenser 1-5 mL
Brinkmann ChemSaver Dispenser 2-10 mL
Brinkmann ChemSaver Dispenser 5-25 mL
Brinkmann ChemSaver Dispenser 10-50 mL.
More Details
Hand Pressure Pumps
Buy Hand Pressure Pumps Items Including:
Hand-operated vacuum/pressure pump, 15 mL/stroke, zinc alloy body
Repair kit for vacuum/pressure pump 79301-11
Hand-operated vacuum pump (without gauge), 32 mL/stroke, PVC body
Low pressure test pump, generates -12 psi/-0.85 bar or pressures to 30.
More Details
Heat Gun Kits
Buy Heat Gun Kits Items Including:
Replacement heating element kit for heat gun 03020-00 (HAS-042K)
Replacement heating element kit for heat gun 03021-00 (HAS-012K)
Replacement heating element kit for heat gun 03022-00 (HAS-041K)
Heat Element Kit for Heat Gun 03033-00 (35031).
More Details
Cole-Parmer Safety Products
Buy Cole-Parmer Safety Products Items Including:
Safety Sign, Danger Lockout Equipment Before Entering, 7 X 10, Aluminu (MLKT106VA)
Safety Sign, Danger Lockout Equipment Before Servicing, 7 X 10, Adhesi (MLKT007VS)
Safety Sign, Danger Lockout Equipment Before Servicing, 7 X 10, Plasti (MLKT007VP).
More Details
High Pressure Hoses
Buy High Pressure Hoses Items Including:
316 Stainless Steel Tubing, high pressure; 1/16" OD; two 3-ft sections (YO-03300-02).
More Details
Vacuum Condenser
Buy Vacuum Condenser Items Including:
Evaporator valve unit for vacuum controller (YO-23123-57)
Woulff Bottle for vacuum controller (YO-23123-58)
Vacuum seal for Buchi Rotary Evaporators (YO-23123-60)
Rotary Evaporator with Diagonal glassware, vacuum regulator, vapor tem (YO-28623-00).
More Details
Usb Thermal Camera
Buy Usb Thermal Camera Items Including:
3 Prime X USB Gradient Upgrade
Flir I40 Thermal Imaging Infrared Camera For Industrial Industry (FLIR I40)
Flir I50 Thermal Imaging Infrared Camera For Industrial Industry (FLIR I50)
Flir T200 Thermal Imaging Infrared Camera (30,000 Pixels) (T200).
More Details
Safety Blade
Buy Safety Blade Items Including:
Disposable safety scapel, SS, #10 blade (YO-10918-00)
Disposable safety scapel, SS, #11 blade (YO-10918-02)
Disposable safety scapel, SS, #12 blade (YO-10918-03)
Disposable safety scapel, SS, #15C blade (YO-10918-05).
More Details
Push in Tube Fittings
Buy Push in Tube Fittings Items Including:
John Guest Push-to-Connect Collet Cover, 5/16" Tube OD
Push-to-connect soft tube supports, 3/8" stem x 5/16" tube OD, 10/pack (YO-34007-38)
Push-to-connect soft tube supports, 1/2" stem x 3/8" tube OD, 10/pack (YO-34007-39).
More Details
Brand Burette
Buy Brand Burette Items Including:
Econ Burette 25 ml 2pk (2116-25)
Econ Burette 25 ml Body Only 6pk (2116-25BO)
Econ Burette Locking Stopcock (2116-LSO)
Pyrex Large Vol Burette 250 ml (2094-250)
Pyrex Large Vol Burette 500 ml (2094-500)
More Details
Bearing Vibration Measurement
Buy Bearing Vibration Measurement Items Including:
Bearing Checker Model BC100
Headphones for Bearing Checker
Measuring Cable, Lemo to BNC Slip-on, 1.5 M
Protective Belt Clip with Wrist Strap and Belt Clip for Bearing Checke
Transducer with quick connectors for adapters
More Details
Perforated Strainer for 2
Buy Perforated Strainer for 2 Items Including:
Sanitary In-Line Strainer Assembly; 2" Tri-Clamp; with 1/8" Perforated.
More Details
Cole-Parmer Oakton Industrial Probes and Thermowells
Buy Cole-Parmer Oakton Industrial Probes and Thermowells Items Including:
Industrial Thermocouple Element, 4"L, 20 Gauge, K type (5504A-K-20-4-0)
Industrial Thermocouple Element, 12"L, 20 Gauge, K type (5504A-K-20-12-0)
Industrial Thermocouple Element, 12"L, 20 Gauge, J type (5504A-J-20-12-0).
More Details
Compressor Air Pressure
Buy Compressor Air Pressure Items Including:
Mid Size Air Filter, 5 micron filter, Max 197 SCFM, Max Pressure 150 P
HD Air Filter, 5 micron filter, Max 288 SCFM, Max Pressure 250 PSIG, 3
HD Air Filter, 5 micron filter, Max 353 SCFM, Max Pressure 250 PSIG, 1
Vacuum/Pressure Pump, 2.0 CFM 115V.
More Details
Straight Scissors
Buy Straight Scissors Items Including:
Iris scissors, Dissecting-grade, Straight (YO-10918-12)
Mayo Dissecting Scissors, Dissecting-grade, Straight (YO-10921-02)
Dissecting scissors, Precision-grade, straight, blunt point
Mayo dissecting scissors, Precision-grade, Straight, blunt point.
More Details
Oakton Self-Adhesive Surface Probes
Buy Oakton Self-Adhesive Surface Probes Items Including:
Type J Self-Adhesive Probe (031804M-J)
Type K Self-Adhesive Probe (031804M-K).
More Details
Fittings for Glass
Buy Fittings for Glass Items Including:
Ring Top Antiroll Thermometer Fittings (D-42)
Triangular Antiroll Thermometer Fittings (D-41)
Snap caps, fits 34534-02, 34534-04 (99500-2)
Reusable screw cap with PTFE liner, fits 34534-06 (9998-13)
Screw cap, fits 34534-06 (99999-13).
More Details
Lcr Circuit
Buy Lcr Circuit Items Including:
886 : Synthesized In-Circuit LCR/ESR meter w/100kHz test frequency (886).
More Details
Red Lion Timers
Buy Red Lion Timers Items Including:
Timer, Red/Green Display Backlight Display.
More Details
Cole-Parmer Digital Timers
Buy Cole-Parmer Digital Timers Items Including:
Cole-Parmer Pocket Timer with 10 Minute Markings & Magnet on Back (YO-94415-44)
Cole-Parmer Digital Lab-Top Alarm Timer
Cole-Parmer Digital Water-Resistant Flashing Timer
Cole-Parmer Digital Tie Timer
Cole-Parmer Continuous Visual Alarm Timer.
More Details
Cole-Parmer Bailers
Buy Cole-Parmer Bailers Items Including:
Disposable bailer, PVC, 1-2/3" OD x 3' (BPWSU1.66X36)
Standard, transparent, reusable, PVC bailer, 1.66" OD x 3' L (BPCS1.66X36)
Slow Emptying Device, for 0.5" OD PE Bailers (YO-99157-22)
Polyester line, 328 ft (100 M) long (62-C50).
More Details
Current Clamp
Buy Current Clamp Items Including:
AEMC CM605 Low Current Clamp Meter, 100 A AC/DC (CM605)
Amprobe Model AC50A, Leakage Current Clamp Multimeter (AC50A)
Extech MA410 400 A AC Current Clamp Meter with Built-In Non-Contact Vo (MA410)
I1A/10A Mini Current Clamp Set (I1A/10A Clamp PQ3).
More Details
Us Safety Goggles
Buy Us Safety Goggles Items Including:
Uvex STEALTH Safety Goggles, Clear Lens
Uvex FUTURA Safety Goggles
Laser Goggles, YAG/KTP
Allsafe MonoGoggle, Regular, Clear Lens
Allsafe MonoGoggle, Large, Clear Lens
More Details
Db Meter Software
Buy Db Meter Software Items Including:
Casella CEL-620.A1/K2 Type 1 Sound Meter Kit with Software
Casella CEL-630.A2/K2 Type 2 Sound Meter Kit with Software
Casella CEL-630.C2/K2 Type 2 Third Octave Sound Meter Kit with Softwar
Casella CEL-620.A2/K2 Type 2 Sound Meter Kit with Software.
More Details
Din Relays
Buy Din Relays Items Including:
Temperature Controller, TC, 1/16-DIN, Relay Output, 2 alarms (DC1010CT-102-000-E)
Temperature Controller, RTD, 1/16-DIN, Relay Output, 1 alarm (DC1014CR-101-000-E)
Temperature Controller, RTD, 1/4-DIN, Relay Output, 1 alarms, (DC1044CR-101-000-E).
More Details
Reusable Ear Plugs
Buy Reusable Ear Plugs Items Including:
Howard Leight Reusable Quiet™ Earplugs with 12" cord (QD30)
AIRSoft™ Reusable Earplugs with cord.
More Details
Cole-Parmer Ptfe Filters
Buy Cole-Parmer Ptfe Filters Items Including:
Cole-Parmer PTFE Nonsterile Syringe Filters; 0.45µ (YO-02915-22)
Cole-Parmer PTFE Nonsterile Syringe Filters; pore size; 0.20µ
Cole-Parmer PTFE Nonsterile Syringe Filters; pore size; 0.45µ
Cole-Parmer PTFE Sterile Syringe Filters; pore size; 0.45µ.
More Details
70618
results
<<<Previous
1
2
3
4
5
6
7
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Ion Filter to Cole-Parmer Ptfe Filters
>
31125329-s-toc_2 31125329stoc2