Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 30

Daq Module   to   Ansell Hyflex

Image
  Product Detail
More Detail

Daq Module

Buy Daq Module Items Including:
  • IOtech Personal Daq 3005™ USB, 1-MHz, 16-bit Multifunction Data Acq
  • IOtech Personal Daq 3000™ USB, 1-MHz, 16-bit Multifunction Data Acq
  • IOtech Personal Daq 3001™ USB, 1-MHz, 16-bit Multifunction Data Ac.

Aurelia Gloves

Buy Aurelia Gloves Items Including:
  • AURELIA Honey-Comb™ Textured Latex Gloves, small
  • AURELIA Honey-Comb™ Textured Latex Gloves, Medium
  • AURELIA Honey-Comb™ Textured Latex Gloves, large
  • AURELIA Honey-Comb™ Textured Latex Gloves, x-large
  • AURELIA Robust™ Nitrile Gloves, Medium, 100/Box.

Tap Fittings

Buy Tap Fittings Items Including:
  • Metalex Paddle-Wheel Flow Sensors, for use with mini-tap fitting; 1-1/ (P525-1)
  • P5262015:Mtlx Minitap1.5in Metalex Mini-Tap (P526-2015)
  • P5262020:Mtlx Minitap2.0in Metalex Mini-Tap (P526-2020)
  • P5262030:Mtlx Minitap3.0in Metalex Mini-Tap (P526-2030).

Safety Step

Buy Safety Step Items Including:
  • Safety Sign, Caution - Watch Your Step, 7" X 10", Adhesive Vinyl
  • Safety Sign, Caution - Watch Your Step, 7" X 10", Aluminum
  • Safety Sign, Caution - Watch Your Step, 10" X 14", Adhesive Vinyl.

H-B Instruments Hydrometers

Buy H-B Instruments Hydrometers Items Including:
  • H-B Instrument Hydrometer, —1 to 12° Baume, 0.1° Divisi
  • H-B Instrument Hydrometer, —49 to 61° Baume, 0.1° Divis
  • H-B Instrument Hydrometer, —0 to 75° Baume, 0.5° Divisi
  • H-B Instrument Hydrometer, 1.000 to 1.225 Specific Gravity, 0.005 Divi.

Bel-Art Bacteria Test Kits

Buy Bel-Art Bacteria Test Kits Items Including:
  • RAMBACH agar, isolation media, 4 x 1L (YO-36152-76)
  • SMAC agar, 500g (YO-36152-26)
  • CT-supplement, 16 vials (YO-36152-28)
  • Listeria selective enrichment supplement, 16 vials (YO-36152-44)
  • OXFORD Listeria selective supplement, 13 vials (YO-36152-52).

Glass Vials with Caps

Buy Glass Vials with Caps Items Including:
  • Glass Vials PTFE Lined Cap Clear 12 ml 200/pk (B7800-12)
  • Glass Vials PTFE Lined Cap Clear 16 ml 200/pk (B7800-4)
  • Glass Vials PTFE Lined Cap Clear 20 ml 100/pk (B7800-20)
  • Glass Vials PTFE Lined Cap Clear 22 ml 200/pk (B7800-5).

Cole-Parmer Starter Units

Buy Cole-Parmer Starter Units Items Including:
  • Adjustable four-shelf starter unit; 36" x 18" x 64" (A1836CP64S-4)
  • Adjustable four-shelf starter unit; 48" x 18" x 64" (A1848CP64S-4)
  • Adjustable five-shelf starter unit; 36" x 24" x 84" (A2436CP84S-5).

Cole-Parmer Shop Coat

Buy Cole-Parmer Shop Coat Items Including:
  • Shop coat; size, small (36-38)
  • Shop coat; size, medium (40-42)
  • Shop coat; size, large (44-46).

Cole-Parmer Ribbed Matting

Buy Cole-Parmer Ribbed Matting Items Including:
  • Ribbed low-density polyethylene matting, 15' long (404225-0003).

Pressure Metering Valve

Buy Pressure Metering Valve Items Including:
  • Economical High-Pressure Single-Piston Pump; 8.0 mL/min Max Flow, 230 (B-100-S-2-CE (#1442))
  • High-pressure piston pump, single-piston, 0.5 to 5.0 mL/min, 1000 psi, (1384)
  • High-pressure piston pump, single-piston, 0.5 to 5.0 mL/min, 1000 psi, (1385).

Calipers and Micrometers

Buy Calipers and Micrometers Items Including:
  • NIST-Traceable Calibration; Micrometers/Calipers, 0-12" (YO-17050-12)
  • NIST-Traceable Calibration; Micrometers/Calipers, 0-24" (YO-17050-13)
  • NIST-Traceable Calibration; Height Gauge, 0-24" (YO-17050-14).

Temp Data Logger

Buy Temp Data Logger Items Including:
  • Extech 42265 : Temp Datalogger Kit W/Pc Interface.

Circuit Breaker Lockouts

Buy Circuit Breaker Lockouts Items Including:
  • Circuit Breaker lockouts; single-pole breakers, 6/pack
  • Single-Pole No Hole Circuit Breaker Lockout (KDD123)
  • Double-Triple Pole Circuit Breaker Lockout (KDD131)
  • Grip Tight, Single/Double Pole Circuit Breaker Lockout (KDD135).

Filter Air Conditioner

Buy Filter Air Conditioner Items Including:
  • Foam Filter For 3302xxx, 3303xxx (YO-30002-33)
  • Foam Filter For 3304xxx, 3305xxx, 3328xxx, 3329xxx, 3332xxx (YO-30002-34)
  • Metal Filter For 3302xxx, 3303xxx (YO-30002-36)
  • Metal Filter For 3304xxx, 3305xxx, 3328xxx, 3329xxx, 3332xxx (YO-30002-37).

Pressure Chart Recorders

Buy Pressure Chart Recorders Items Including:
  • 8 IN Circular Chart Recorder - Pressure, 1 pen, Flush/back mount (YO-30018-13)
  • 8 IN Circular Chart Recorder - Pressure, 1 pen, Pipestand/bottom mount (YO-30018-15)
  • 8 IN Circular Chart Recorder - Pressure, 2 pen, Pipestand/bottom mount (YO-30018-16).

Diaphragm Seals

Buy Diaphragm Seals Items Including:
  • In-line Seal, 1 1/2" Process Conn, w/ 15 PSI Transmitter
  • In-line Seal, 1 1/2" Process Conn, w/ 30 PSI Transmitter
  • In-line Seal, 1 1/2" Process Conn, w/ 100 PSI Transmitter
  • In-line Seal, 2" Process Conn, w/ 15 PSI Transmitter.

Clock Humidity

Buy Clock Humidity Items Including:
  • Cole-Parmer Thermometer/Clock/Humidity Thermometer
  • Digital Termperature and Humidity Monitor with Clock and Alarm
  • Extech 445702 Thermohygrometer Clock (445702)
  • Thermohygrometer Wall Clock (7161CP)
  • Large digital clock with humidity and temperature.

Cell Culture Products

Buy Cell Culture Products Items Including:
  • Corning cell culture dish, non-treated, 60 mm diameter, 500/cs
  • Systems for cell culture applications, 115 VAC (YO-29200-12)
  • Systems for cell culture applications, 220 VAC (YO-29200-13)
  • Systems for cell culture applications, 115 VAC (YO-29200-32).

Small Cord

Buy Small Cord Items Including:
  • Flexible, small-diameter, fiberglass insulated heating cord, 3', 53 wa (YO-03122-30)
  • Line Cord (US - 115V) for use with Cole-Parmer Syringe Pump, Infusion (788308) (YO-74903-78).

Stainless Steel Static Mixer

Buy Stainless Steel Static Mixer Items Including:
  • Pipe Mixer, 316 stainless steel, 1/2" NPT(M), 12 elements (YO-04669-18)
  • Pipe Mixer, 316 stainless steel, 3/4" NPT(M), 12 elements (YO-04669-20)
  • Pipe Mixer, 316 stainless steel, 1-1/2" NPT(M), 6 elements (YO-04669-23).

Rubber Stoppers

Buy Rubber Stoppers Items Including:
  • Black rubber stopper, size 00
  • Black rubber stopper, size 12
  • Black rubber stopper, size 13
  • Black rubber stopper, size 14
  • Black rubber stopper, size 15

Aluminum 100

Buy Aluminum 100 Items Including:
  • General Purpose Aluminum Weighing Dishes, 44mm i.d., 51mm o.d
  • x 1/2" (D44-100)
  • General Purpose Aluminum Weighing Dishes, 63mm i.d., 71mm o.d
  • x 11/16 (D44-100)
  • General Purpose Aluminum Weighing Dishes, 62mm i.d., 70mm o.d

Cole-Parmer Environmental Testing

Buy Cole-Parmer Environmental Testing Items Including:
  • Water, ECD tested for pesticide analysis (2.5l) (YO-88294-46)
  • Wet/dry soil sampling tube 18" long (PN013)
  • T-handle (PN016)
  • Clay soil auger, carbon steel, 3' diameter (PN070)
  • Soil auger handgrip (PN025)

Microscope Ccd

Buy Microscope Ccd Items Including:
  • Digital 2300 CCD microscope camera with software (SP100382D MOTICAM 2300)
  • Motic Moticam Pro Digital Microscope Camera, 1/2" CCD, Peltier cooled
  • Motic Moticam Pro Digital Microscope Camera, 1/1.8" CCD, Peltier coole.

Relative Humidity Transmitters

Buy Relative Humidity Transmitters Items Including:
  • Vaisala Relative Humidity Transmitter, Duct-Mount (HMD60U)
  • GE (General Eastern) Industrial relative Humidity/Dew Point Analyzer, (MR2350) (YO-37405-04)
  • Vaisala Relative Humidity And Temperature Transmitter (no Display - Fi (HMT100-A02A122A2A1AB00).

Force Load Cells

Buy Force Load Cells Items Including:
  • 22 lb capacity smart "S" type load cell
  • 110 lb capacity smart "S" type load cell
  • 1100 lb capacity smart "S" type load cell
  • Load Cell, 10 lb
  • Load Cell, 20 lb

Cole-Parmer Pure Polypropylene Housing and Sump

Buy Cole-Parmer Pure Polypropylene Housing and Sump Items Including:
  • Double open end polypropylene housing and sump, 10" (150132).

Tube 12

Buy Tube 12 Items Including:
  • 166-12 Stainless Steel Pitot Tube 12" Insertion Length 1/8" Diameter
  • Microbore - Puri-Flex ™ Autoanalysis Tubing, -12, 50 feet
  • Wet/dry soil sampling tube 12" long (PN012)
  • Thermo Scientific Nunc Round-bottom tubes; 12-mL; PC; 10/pack.

Ansell Hyflex

Buy Ansell Hyflex Items Including:
  • Ansell HyFlex™ Work Gloves, Size 7
  • Ansell HyFlex™ Lite Work Gloves, Size 7
  • Ansell HyFlex™ Lite Work Gloves, Size 10
  • Ansell® HyFlex®Cut Resistant Dyneema® Work Gloves, Size 10 (288738)
  • Ansell® HyFlex® Nylon Kit Work Gloves with Nitrile AGT, Size 7.



Daq Module to Ansell Hyflex

>