Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 300

Oakton Ph   to   Esd Tweezers

Image
  Product Detail
More Detail

Oakton Ph/Ion Buffers, and Storage Solutions

Buy Oakton Ph/Ion Buffers, and Storage Solutions Items Including:
  • Oakton electrode storage solution
  • Oakton pH Solution Pack, one 500-mL bottle each of pH 4.01, 7.00, 10.0
  • Oakton electrode storage solution, case of 24
  • Oakton electrode cleaning solution
  • Oakton 4.01 pH buffer, 500-mL

Water Analysis Kits

Buy Water Analysis Kits Items Including:
  • Refill kit, Phenol test, for 00290-30
  • Refill kit, Ammonia/nitrogen, Nesslerization test, for 00291-52, six-m
  • Refill kit, Iron, total soluble test, for 00291-40
  • DO Probe Service Kit for Hach DO Probes
  • DO probe maintenance kit for DO probe 53101-51.

Cole-Parmer Pyridine-D5

Buy Cole-Parmer Pyridine-D5 Items Including:
  • PYRIDINE-D5 99% ATOM D 10ML (YO-88232-31).

Split Core Current Transducers

Buy Split Core Current Transducers Items Including:
  • CR4120S-5 Split-Core 4-20mA/0-5 Amp
  • CR5210S-300, +/- 5 VDC output, DC Current, 0-300Amps
  • CR4110S-15, 0 - 5 VDC output, AC True RMS, 0-15 Amps
  • CR4110S-50, 0 - 5 VDC output, AC True RMS
  • 0-50 Amps

10 Bottle

Buy 10 Bottle Items Including:
  • Mark-10 MR53-10Z Torque Sensor, 10 oz x 700 g cm x 7 N cm
  • Mark-10 MR53-20Z Torque Sensor, 20 oz x 1400 g cm x 14 N cm
  • Mark-10 MR53-100 Torque Sensor, 100 lb x 1600 oz in x 1150 N cm
  • Thermo Scientific Nalgene polycarbonate carboy with spigot, 10 L.

Cole-Parmer Non-Sealed Tub Assemblies

Buy Cole-Parmer Non-Sealed Tub Assemblies Items Including:
  • Tub assembly, non-sealed, 5 L, with port for temperature/humidity prob (50035010)
  • Tub assembly, non sealed, 2 L (50035005)
  • Tub assembly, non-sealed 2 L (50035006)
  • Tub assembly, non-sealed, 5 L (50035008).

Rice Lake Weighing

Buy Rice Lake Weighing Items Including:
  • Rice Lake Weighing Systems Calibration Weight, Class 4 with Traceable (80505TR)
  • Rice Lake Weighing Systems Calibration Weight, Class 4 with Traceable (80504TR)
  • Rice Lake Weighing Systems Calibration Weight, Class 4 with Traceable (80502TR).

Three Way Check Valve

Buy Three Way Check Valve Items Including:
  • PVDF Three-Way Stopcock, 6 mm hose barb, 4 mm orifice
  • PVDF Three-Way Stopcock, 8 mm hose barb, 6 mm orifice.

Abb Commander Circular Chart Recorders

Buy Abb Commander Circular Chart Recorders Items Including:
  • ABB Commander Circular Chart Recorder/Controller - 1 controller, 2 pen
  • Chart Paper for ABB Commander Circular Chart Recorder/Controllers, 0 t (500P1225-106)
  • Chart Paper for ABB Commander Circular Chart Recorder/Controllers, 0 t (500P1225-15).

Cole-Parmer Right-to-Know Compliance Centers

Buy Cole-Parmer Right-to-Know Compliance Centers Items Including:
  • Right-To-Know Compliance Center, Single binder, English/Spanish (2010SP)
  • Right-To-Know Compliance Center, Double binders, English (2010DB)
  • Replacement MSDS Binder, 1 1/2", English (2023)
  • Replacement MSDS Binder, 1-1/2", English/Spanish.

Cole-Parmer Cleanroom Labels

Buy Cole-Parmer Cleanroom Labels Items Including:
  • Cleanroom Labels, Removable, 2" x 2".

Plus Digital

Buy Plus Digital Items Including:
  • Accu-cal Digital Test Gauge, Vac To 15 Psi (75514-15B55)
  • Accu-cal Digital Test Gauge, Vac To 15 Psi (75514-15B15)
  • Accu-cal Digital Test Gauge, Vac To 1000 Psi, W/ Flange
  • Accu-cal Digital Test Gauge, Vac To 2000 Psi, W/ Flange.

Pressure Sensor Transmitter

Buy Pressure Sensor Transmitter Items Including:
  • PendoTECH PressureMAT Monitor/Transmitter, Four Pressure Sensor Inputs
  • TB-2012M : Barometric Pressure Sensor Analog 4-20mA
  • Diaphragm Sensor, 1-1500 Torr, 1/2" O.d..

Replacement Motor 115v

Buy Replacement Motor 115v Items Including:
  • Replacement 600 rpm, 115V motor for 07552-70 Masterflex L/S system.
  • Replacement 600 rpm, 115V motor for Masterflex L/S systems..

Steel Beams

Buy Steel Beams Items Including:
  • Ohaus Triple-Beam Balance, 610 g x 0.1 g, Stainless Steel weighing pla (760-00)
  • Ohaus Dial-O-Gram Triple Beam Balance with Tare and Stainless Steel Pa
  • Ohaus Harvard Triple Balance, 2000g X 0.1g, two beam, Stainless Steel (YO-11110-21).

Cabinet Air Conditioners

Buy Cabinet Air Conditioners Items Including:
  • TS8 Roof Plate for air conditioner 24X24
  • AC With Condensate Evaporator 37.4" X 15.7" X 10.2", 2.6amps, 400v 3-5 (YO-30002-20)
  • AC With Condensate Evaporator 37.4" X 15.7" X 10.2", 12.5amps, 230V (YO-30002-21).

Burrell Shakers

Buy Burrell Shakers Items Including:
  • Wrist-Action Shaker, 8 Flask, 115VAC, 60Hz
  • Wrist-Action Shaker, 8 Flask, 230VAC, 50Hz
  • Wrist-Action Shaker, 12 Flask, 115VAC, 60Hz
  • Wrist-Action Shaker, 12 Flask, 230VAC, 50Hz
  • Wrist-Action Shaker, 4 Flask-Side Arm, 8 Flask Platform, 115VAC, 60Hz.

Silicone Tubing Properties

Buy Silicone Tubing Properties Items Including:
  • Peroxide-Cured Reinforced Silicone Tubing, 175 psi at 70°F, 5/16" (YO-95623-00)
  • Peroxide-Cured Reinforced Silicone Tubing, 175 psi at 70°F, 3/8" ( (YO-95623-01)
  • Peroxide-Cured Reinforced Silicone Tubing, 155 psi at 70°F (YO-95623-03).

Gooch Crucibles

Buy Gooch Crucibles Items Including:
  • CoorsTek® Gooch Filter-Crucible, Porcelain; 13 mL, for 15-16 mm dia fi (YO-17805-00)
  • CoorsTek® Gooch Filter-Crucible, Porcelain; 25 mL, for 20-21 mm dia fi (YO-17805-02)
  • CoorsTek® Gooch Filter-Crucible, Porcelain; 40 mL, for 23-24 mm dia fi.

Colder Hose Barb Inserts

Buy Colder Hose Barb Inserts Items Including:
  • Quick-Disconnect Hose Barb Insert; Valved, 1/16 in
  • Tube ID, 1/8 in
  • F (PMCD2201)
  • Quick-Disconnect Hose Barb Insert; Valved, 1/4 in
  • Tube ID, 1/4 in

Cole-Parmer Mettler-Toledo-Compatible Electrodes

Buy Cole-Parmer Mettler-Toledo-Compatible Electrodes Items Including:
  • Double junction electrode w/ Tuff-Tip (YO-27508-04).

Cole-Parmer Conical-Bottom Tank Systems

Buy Cole-Parmer Conical-Bottom Tank Systems Items Including:
  • Conical-Bottom Tank System, 100 Gal
  • (YO-06925-15)
  • Open Top Tank, conical bottom 120GAL CONE BOTTOM (YO-06960-60)
  • Open Top Tank, conical bottom 150GAL CONE BOTTOM (YO-06960-62)
  • Open Top Tank, conical bottom 200GAL CONE BOTTOM (YO-06960-64).

Thermo Scientific Orion Ph/Ion Handheld Meters

Buy Thermo Scientific Orion Ph/Ion Handheld Meters Items Including:
  • Thermo Scientific Orion Star A121 pH Portable Meter Only
  • Thermo Scientific Orion Star A324 pH/ISE Portable Meter Only
  • Thermo Scientific Orion Star A324 pH/ISE Portable Meter Kit
  • Thermo Scientific Orion Star A325 pH/Cond Portable Meter Kit.

Whatman Qualitative Filter Papers

Buy Whatman Qualitative Filter Papers Items Including:
  • Whatman Qualitative Filter Papers; 5.5 cm dia; pore size, 20-25µ; 1
  • Whatman Qualitative Filter Papers; 11.0 cm dia; pore size, 20-25 µ;
  • Whatman Qualitative Filter Papers; 12.5 cm dia; pore size, 20-25 µ;.

Salisbury Arc Flash

Buy Salisbury Arc Flash Items Including:
  • Salisbury Pro-Wear Arc Flash Jackets, 8 cal/cm2, Extra Large
  • Salisbury Pro-Wear™ Arc Flash Protective Overpants, 8 cal/cm2, Medi
  • Insulated Glove, Class 0, Black, 11" length, size 8-1/2
  • Insulated Glove, Class 0, Black, 11" length, size 10.

Pp Bottles

Buy Pp Bottles Items Including:
  • Thermo Scientific Nalgene polypropylene narrow-mouth bottle, 1/8 oz
  • Thermo Scientific Nalgene polypropylene narrow-mouth bottle, 1/4 oz
  • Thermo Scientific Nalgene polypropylene narrow-mouth bottle, 1/2 oz
  • 4 Liter PP bottle.

Microfluidic Applications

Buy Microfluidic Applications Items Including:
  • Microfluidic Fluid Splitter Slide, Several splitters on one slide
  • Microfluidic Snake Mixer Slide, COC material, several mixers on one ch
  • Microfluidic Microplat Assembly Platform
  • Microfluidic Micro Luer Connector, COC material, package of 25 pieces.

Oakton Economical Ptfe-Coated Probes

Buy Oakton Economical Ptfe-Coated Probes Items Including:
  • Low-Cost RTD PTFE-coated probe; 4.5"L (111897C)
  • Type J Low-cost PTFE Thermocouple Probe, 4.5"L (070697A-J)
  • Type K Low-cost PTFE Thermocouple Probe;4.5"L (070697A-K)
  • Type T Low-cost PTFE Thermocouple Probe; 4.5"L (070697A-T).

Spirax Sarco Bronze Y Stainers

Buy Spirax Sarco Bronze Y Stainers Items Including:
  • Bronze "Y" strainer, 1/4" NPT(F) (BT 1/4" 51988 )
  • Bronze "Y" strainer, 3/8" NPT(F) (BT 3/8" 51989 )
  • Bronze "Y" strainer, 1/2" NPT(F) (BT 1/2" 51990 ).

Esd Tweezers

Buy Esd Tweezers Items Including:
  • ESD Plastic Tweezers, Flat Round Tip, Pattern 702A
  • ESD Plastic Tweezers, Ultra Fine Tip, Pattern 705
  • ESD Plastic Tweezers, Flat Fine Tip, Pattern 709
  • ESD Plastic Tweezers, Flat Wide Tip, Pattern 710
  • ESD Plastic Tweezers, Fine Tip, Pattern 707.



Oakton Ph to Esd Tweezers

>