Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 4
Texas Instruments
to
Short Rod
Image
Product Detail
More Detail
Texas Instruments
Buy Texas Instruments Items Including:
Texas Instruments Mini-Desktop Calculator (TI-1795 SV)
Texas Instruments Two-Line Scientific Calculator (TI-30X2S)
Texas Instruments Scientific Calculator, Solar Powered
Texas Instruments Graphing Calculator with Advanced Statistics (TI-83+).
More Details
Scientific Conversion Calculator
Buy Scientific Conversion Calculator Items Including:
Cole-Parmer Compact Metric Conversion Calculator
Solar-Powered Scientific Calculator (YO-17300-02)
Two line display scientific calculator (CSO-FX-300MS)
Texas Instruments Two-Line Scientific Calculator (TI-30X2S)
Cole-Parmer Scientific Calculator.
More Details
Torque and Angle
Buy Torque and Angle Items Including:
Torque and Angle Electronic Torque Wrench, 10 to 100 ft
lb.
Torque and Angle Electronic Torque Wrench, 25 to 250 ft
lb..
More Details
External Remote
Buy External Remote Items Including:
VAISALA RH Only transmitter w/Display, 5 Meter Remote Probe and 4-20mA
VAISALA RH-temperature transmitter w/Display, 3 Meter Remote Probe and
VAISALA RH-temperature transmitter w/Display, 5 Meter Remote Probe and.
More Details
96 Well Plate Reader
Buy 96 Well Plate Reader Items Including:
96-well Deep Well Plate, Polypropylene, 48/pack (YO-07904-02)
96-well Deep Well Plate, Polypropylene, 24/pack (YO-07904-04)
96-well Deep Well Plate, Polystyrene, 32/pack (YO-07904-06)
Costar 96-Well Standard Microplates, Nontreated, PP, Flat Well.
More Details
Disposable Aluminum Pans
Buy Disposable Aluminum Pans Items Including:
ErgoClip ® accessory kit; includes 10 disposable aluminum pans, 50 we (11106707).
More Details
Laboratory Mortar and Pestle
Buy Laboratory Mortar and Pestle Items Including:
Cole-Parmer Mortar and Pestle Set, Porcelain; 80ML, Each
Cole-Parmer Mortar and Pestle Set, Porcelain; 500ML Each
Mortar and Pestle; Agate, 125ML, Each
Agate Mortar and Pestle, 25 mL (YO-04010-20)
Glass Mortar & Pestle Set, 50ml (YO-34500-51).
More Details
Cole-Parmer Tee Fittings
Buy Cole-Parmer Tee Fittings Items Including:
Cole-Parmer, Sanitary Clamp Tee, 304L SS, 1"
Cole-Parmer, Sanitary Clamp Tee, 304L SS, 1-1/2"
Flow Cell, CPVC tee fitting, 1-1/2" Slip Flit (YO-27113-40)
Female threaded tee fittings, nylon, 1/8" NPT(F), pack of 6 (YO-06349-50).
More Details
Cole-Parmer Hot-Wire Thermoanemometer
Buy Cole-Parmer Hot-Wire Thermoanemometer Items Including:
Cole-Parmer Heavy-Duty Hot-Wire Thermoanemometer with Datalogger.
More Details
Centrifuge Motor
Buy Centrifuge Motor Items Including:
CINC V2 Explosion-Proof Centrifugal Separator/Extractor, 220 VAC
Magnetic Drive Pump with 24 VDC Motor, Glass-Fiber Reinforced Polyprop (YO-72008-00)
Magnetic Drive Pump with 24 VDC Motor, Glass-Fiber Reinforced Polyprop (YO-72008-10).
More Details
Cole-Parmer Polypropylene Sanitary Clamp Concentric Reducers
Buy Cole-Parmer Polypropylene Sanitary Clamp Concentric Reducers Items Including:
Polypropylene Sanitary Concentric Reducer,1.5X3/4" (YO-31805-06)
Polypropylene Sanitary Concentric Reducer, 2" x 1" Ladish Clamp Size (YO-31805-11)
Polypropylene Sanitary Concentric Reducer, 3" x 2" Clamp Size (YO-31805-18).
More Details
Cole-Parmer Round Panel Jacks - Standard
Buy Cole-Parmer Round Panel Jacks - Standard Items Including:
Type J Round Standard panel jack; color, black (H32J)
Type K Round Standard panel jack; color, yellow (H32K).
More Details
Litmus Test Paper
Buy Litmus Test Paper Items Including:
Litmus Blue Test Paper, 10 Booklets of 20 Strips.
More Details
Vibration Mount
Buy Vibration Mount Items Including:
Vibrasorb Mount, Large
Vibrasorb Mount, Small
Vibrasorb Damping Mount
Flowline Switch-Pak Vibration Sensor PP Wetted Material Controller W/
Flowline Switch-Pak Vibration Sensor PP Wetted Material Controller On.
More Details
Centrifugal Submersible Pumps
Buy Centrifugal Submersible Pumps Items Including:
Economical Submersible Pump, High-Flow Utility, 20.4 GPM, 115 VAC, 18'
Economical Submersible Pump, High-Flow Utility, 20.4 GPM, 230 VAC, 12'
Submersible Pump for Corrosive/Acid Sumps, 304 SS Metal Fitted, Buna N.
More Details
Drawers and Shelves
Buy Drawers and Shelves Items Including:
SS equipment cart with drawers; 18-3/8" x 16-5/8", 3 shelves, 200 lb c (356)
SS equipment cart with drawers; 18-3/8" x 16-5/8", 2 shelves, 200 lb c (358).
More Details
Tube Flowmeter
Buy Tube Flowmeter Items Including:
Pocket Flowmeter for Oxygen, 1 to 8 LPM; Fits 3/16 to 7/16" ID Tubing
Polyproylene Tube Holder for 8.5 mm PFA and PVDF Flow Sensors
Replacement 4.5 mm PVDF Turbine Flow Sensor for PP Tube Holder 32516-3
Gilmont Unshielded Flowmeter Sz
0 Tube.
More Details
Temperature Control Systems
Buy Temperature Control Systems Items Including:
Mass flow controller, universal COMM, for Air/N2/CO, 0 to 1 (GFCS-011293)
Mass flow controller, universal COMM, for Air/N2/CO, 0 to 1 (GFCS-011545)
Mass flow controller, universal COMM, for Air/N2/CO, 0 to 2 (AFCS-010001).
More Details
Advantec Filter Membranes
Buy Advantec Filter Membranes Items Including:
Advantec Sterile Mixed Cellulose Ester Filter Membranes; 47mm dia; 0.6 (YO-06644-06)
Advantec Sterile Mixed Cellulose Ester Filter Membranes; 47mm dia; 0.4 (YO-06644-08).
More Details
Lab Shelving
Buy Lab Shelving Items Including:
Electronic Desiccator with Flexboard Shelves; interior dim., 11-1/2"W (134041-2)
Electronic Desiccator with Stainless Steel Shelves; interior dim.,11-1 (134241)
Electronic Desiccator with Stainless Steel Shelves; interior dim., 11- (134241-2).
More Details
Allegro Low-Pressure Supplied-Air Respirator Systems
Buy Allegro Low-Pressure Supplied-Air Respirator Systems Items Including:
Replacement Tyvek Hood for Supplied Air Systems
Allegro One Man Full Face Piece Supplied Air System with 50 foot Hose
Allegro One Man Tyvek Hood Supplied Air System with 50 foot Hose
Allegro Two Man Tyvek Hood Supplied Air System with 50 foot Hose.
More Details
Precision General Purpose Incubators
Buy Precision General Purpose Incubators Items Including:
High-performance mechanical convection incubator, 2 doors, 11.2ft (PR205075M)
High-performance mechanical convection incubator, 2 doors, 11.2ft (PR205070M)
High-performance 2-door gravity convection incubator, 11.2ft3/su (PR205075G).
More Details
Lamotte Spectroscopy and Spectrophotometers
Buy Lamotte Spectroscopy and Spectrophotometers Items Including:
Aluminum test kit (3641-SC)
Ammonia nitrogen test kit (365901-SC)
Chlorine test kit (6739-G)
Fluoride test kit (3647-01-SC)
Iron test kit (3668-SC)
More Details
Three Phase Inverter
Buy Three Phase Inverter Items Including:
AC Drive / Frequency Inverter, 200-240 VAC single or three phase input (ESV251N02SXB).
More Details
Master Recorder
Buy Master Recorder Items Including:
AC-powered, Modbus master & EtherNet/IP Client/Server, one logic and t
AC-powered, Modbus master & EtherNet/IP Client/Server, one logic, one
AC-powered, two PID loops, dual setpoint programmer, Modbus master & E (YO-70002-67).
More Details
Thermo Scientific Nalgene Cryogenics
Buy Thermo Scientific Nalgene Cryogenics Items Including:
Thermo Scientific Nalgene Cryogenic vial; 1.0 mL, 25/pack
Thermo Scientific Nalgene Cryogenic vial; 1.5 mL, 25/pack
Thermo Scientific Nalgene Cryogenic vial; 1.2 mL, 25/pack
Thermo Scientific Nalgene Standard Cryocane, Aluminum, 300 mL (holds 6.
More Details
Ultrasonic Products Surface Testing
Buy Ultrasonic Products Surface Testing Items Including:
NIST Non-Metal Coating Thickness Gauge, 0.5 - 40 mils
NIST Non-Metal Coating Thickness Gauge, 0.5 - 40 mils, measures up to
NIST Non-Metal Coating Thickness Gauge, 2 - 150 mils
NIST Non-Metal Coating Thickness Gauge, 2 - 150 mils, measures up to 3.
More Details
Light Meter Calibration
Buy Light Meter Calibration Items Including:
Cal-LIGHT Precision 400 Light Meter with back light display, case, bat
Dual-Scale Light Meter with NIST-traceable certificate of calibration (840020C).
More Details
Tri-Clamp Strainers
Buy Tri-Clamp Strainers Items Including:
Sanitary In-Line Strainer Assembly; 1" Tri-Clamp; with 1/8" Perforated
Sanitary In-Line Strainer Assembly; 1.5" Tri-Clamp; with 1/8" Perforat
Sanitary In-Line Strainer Assembly; 2" Tri-Clamp; with 1/8" Perforated.
More Details
Short Rod
Buy Short Rod Items Including:
Immersion heater, short rod, 5.5" L heated area, 500 watts, 115 VAC (324-114)
Immersion heater, short rod, 5.5" L heated area, 500 watts, 230 VAC (306-114)
Immersion heater, short rod, 5" L heated area, 1100 watts, 115 VAC (324-230).
More Details
70618
results
<<<Previous
1
2
3
4
5
6
7
8
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Texas Instruments to Short Rod
>
31125329-s-toc_2 31125329stoc2