Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 40
Kleenpak Fittings
to
Electric Thermometers
Image
Product Detail
More Detail
Kleenpak Fittings
Buy Kleenpak Fittings Items Including:
Kleenpak™ Connector, Polycarbonate, Female, 1/4, pack of 25 (KPCHT02F7)
Kleenpak™ Connector, Polycarbonate, Female, 3/8, pack of 25 (KPCHT02F10)
Kleenpak™ Connector, Polycarbonate, Male, 3/8, pack of 25 (KPCHT02M10).
More Details
Intrinsically Safe Products
Buy Intrinsically Safe Products Items Including:
GE (Druck) Intrinsically Safe Advanced Modular Calibrator with HART Co (DPI 620 IS) (YO-50015-02)
Thermo Scientific Intrinsically Safe Portable Conductivity Meter
Individual Intrinsically Safe micro noise badge, with mounting clips a (CEL-350IS).
More Details
Magnetic Seal
Buy Magnetic Seal Items Including:
Magnetic Drive Pump with 24 VDC Motor, Glass-Fiber Reinforced Polyprop (YO-72008-00)
Magnetic Drive Pump with 24 VDC Motor, Glass-Fiber Reinforced Polyprop (YO-72008-10).
More Details
Digital Timers
Buy Digital Timers Items Including:
Cole-Parmer Extra-Large Digit Timer
Cole-Parmer Pocket Keychain Digital Timer
Cole-Parmer Ultrasonic Cleaner with Digital Timer and Heater, 1/2 gall (YO-08895-27)
Cole-Parmer Ultrasonic Cleaner with Digital Timer and Heater, 1/2 gall (YO-08895-35).
More Details
Chiller Control Temperature
Buy Chiller Control Temperature Items Including:
Cole-Parmer Polystat Temperature-controlled Cooling Recirculators; tem (YO-12910-00)
PolyScience Recirculator; -10 to 70°C; 1 hp; centrifugal pump with (YO-12910-21)
PolyScience Recirculator; -10 to 70°C; 1 hp; centrifugal pump with (YO-12910-22).
More Details
Flashlight Bulb
Buy Flashlight Bulb Items Including:
Maglite® Solitaire Replacement bulb, 2/pk
Mini-Maglite® AAA Replacement bulb, 2/pk
Mini-Maglite® AA Replacement bulb, 2/pk
Maglite® C-cell and D-cell Replacement bulb, 2/pk
Maglite® Mag Charger Rechargeable Flashlight, NiCD batteries, blac.
More Details
Cole-Parmer Mobile Bench Cabinet
Buy Cole-Parmer Mobile Bench Cabinet Items Including:
Mobile laboratory bench, putty (PMB7816).
More Details
Vibration Test
Buy Vibration Test Items Including:
Fluke 810 Vibration Testing System
Low range tuning fork vibration viscometer 120 VAC
High range tuning fork vibration viscometer 120 VAC
High range tuning fork vibration viscometer 240 VAC.
More Details
Unit Mass Flow Controllers
Buy Unit Mass Flow Controllers Items Including:
Flow controller set point module, 100 units/min scale
Flow controller set point module, 200 units/min scale
Flow controller set point module, 250 units/min scale
Flow controller set point module, 500 units/min scale.
More Details
Heat Gun Temperature
Buy Heat Gun Temperature Items Including:
Proheat Variair Heat Gun, 130 to 900°F, 120 V
Economical Heat Gun, 500/1000°F, 120 V
Variable temperature heat gun 72 to 1000°, 1740 watts, 120 VAC, 60 (VT-750C)
Variable temperature heat gun 72 to 750°, 1680 watts, 230 VAC, 50/ (VT-752C-A1).
More Details
Corning Chromatography Columns with Reservoirs
Buy Corning Chromatography Columns with Reservoirs Items Including:
Corning Glass Chromatography Column with Reservoir, 200 mL.
More Details
Ultra Freezers
Buy Ultra Freezers Items Including:
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 14.9 c (YO-44146-03)
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 19.4 c (YO-44146-04)
Thermo Scientific Revco; UxF Series, -86°C Upright Freezer, 19.4 c (YO-44146-05).
More Details
Spill-X-Fp Safety Products
Buy Spill-X-Fp Safety Products Items Including:
SPILL-X-FP/SPILL-X-S Formaldehyde/Solvent Spill Treatment Kit
SPILL-X-FP Formaldehyde Agent Refill, 6 containers.
More Details
Tygon Tubing
Buy Tygon Tubing Items Including:
Masterflex® Tygon® tubing, nonphthlate/non-DEHP, 1.85mm ID, 1
Tygon® high-purity tubing, 1/16"ID x 3/16"OD, 50 ft/pack (YO-95635-00)
TYGON Lab Tubing, 4.0 mm ID, 16 ft/pack
Tygon High Purity Tubing, 1/16"ID x 3/16"OD, 50 ft/pk.
More Details
Chlorine Analyzers
Buy Chlorine Analyzers Items Including:
Free Chlorine Analyzer System, 117 VAC
Free Chlorine Analyzer System, 220 VAC
Fill solution for Free Chlorine sensors, 30-mL bottle (YO-99673-13)
Replacement membrane cap for Free Chlorine sensors (YO-99673-17)
Free Chlorine Replacement Electrode.
More Details
Eppendorf Microcentrifuge
Buy Eppendorf Microcentrifuge Items Including:
Eppendorf® Microcentrifuge Optional Tube Adapter for 0.5-mL and Microt
Eppendorf 5424 Refrigerated Microcentrifuge with 24-tube x 2 mL rotor (YO-02580-01)
Eppendorf 5424 Refrigerated Microcentrifuge with 24-tube x 2 mL rotor (YO-02580-06).
More Details
Desco Anti Static Bags/Esd Bags
Buy Desco Anti Static Bags/Esd Bags Items Including:
Desco Shielding Bag, 10" x 12" (YO-26808-34)
Desco Shielding Bag, 10" x 14" (YO-26808-36)
Desco Shielding Bag, 12" x 16" (YO-26808-38)
Desco Shielding Bag, 15" x 18" (YO-26808-40).
More Details
Eppendorf Fixed-Volume Pipettors
Buy Eppendorf Fixed-Volume Pipettors Items Including:
Ultramicro Fixed-Volume Pipettors, 1 µL (22 47 040-0)
Ultramicro Fixed-Volume Pipettors, 10 µL (22 47 055-8)
Fixed-volume pipettors, 10 µL (22 47 060-4)
Fixed-volume pipettors, 100 µL (22 47 115-5)
Fixed-volume pipettors, 200 µL (22 47 125-2).
More Details
Colored Latex Gloves
Buy Colored Latex Gloves Items Including:
Microflex Color Touch Latex Gloves; pink; size, small
Microflex Color Touch Latex Gloves; green; size, medium.
More Details
Cole-Parmer Relief Valves
Buy Cole-Parmer Relief Valves Items Including:
Liquid Relief Valves 19m 1/2" (0019-C01-MG0020)
Liquid Relief Valves 19m 1/2" (0019-C01-MG0040)
Liquid Relief Valves 20m 1-1/4" (0020-F01-MG0020)
Liquid Relief Valves 20m 1-1/4" (0020-F01-MG0040).
More Details
Chromatography Columns
Buy Chromatography Columns Items Including:
Corning Standard Glass Chromatography Column, 10.5 ID x 150 L
Corning Standard Glass Chromatography Column, 10.5 ID x 250 L
Corning Standard Glass Chromatography Column, 19.0 ID x 250 L
Corning Standard Glass Chromatography Column, 19.0 ID x 500 L.
More Details
Raven Water and Industrial Liquid Sampling
Buy Raven Water and Industrial Liquid Sampling Items Including:
Raven Environmental Products Industrial Process Centrifuge, 110VAC; 60 (F-10300-REDESIGNED)
Raven Environmental Products Industrial Process Centrifuge, 230 VAC; 5 (F-10300-230)
Raven Coretaker Polycarbonate Sludge Sampler, 24-ft (B-10104-24).
More Details
Infrared Scanner
Buy Infrared Scanner Items Including:
Seeker Infrared Thermometer and Scanner with Star Burst Laser Target ( (IRTC50)
Seeker Infrared Thermometer and Scanner with Star Burst Laser Target (.
More Details
Master Appliance
Buy Master Appliance Items Including:
Replacement heating element kit for heat gun 03020-00 (HAS-042K)
Replacement heating element kit for heat gun 03021-00 (HAS-012K)
Replacement heating element kit for heat gun 03022-00 (HAS-041K)
Heat-shrink tubing adapter for heat guns (A-160-HG).
More Details
Techne Laboratory Equipment
Buy Techne Laboratory Equipment Items Including:
Polypropylene balls, 10 mm, pack of 5000 (YO-06821-12)
Polypropylene balls, 20 mm, pack of 500 (YO-06821-22)
Polypropylene balls, 20 mm, pack of 2000 (YO-06821-25).
More Details
Slide-a-Lyzer Dialysis Cassette
Buy Slide-a-Lyzer Dialysis Cassette Items Including:
Thermo Scientific Slide-A-Lyzer Dialysis Cassettes, 3500 MWCO, 0.1-0.5
Thermo Scientific Slide-A-Lyzer Dialysis Cassettes, 3500 MWCO, 0.5-3mL
Thermo Scientific Slide-A-Lyzer Dialysis Cassettes, 3500 MWCO, 3-12mL,.
More Details
Stereo Zoom Microscopes
Buy Stereo Zoom Microscopes Items Including:
Leica Zoom 2000™ Stereozoom Microscope, 10x to 30x Magnification, L
Leica Zoom 2000™ Stereozoom Microscope, 8x to 35x Magnification, LE
Stereozoom Microscope with Five-Setting Illuminator, 10x to 30x zoom, (YO-03904-22).
More Details
Fluke Oscilloscopes
Buy Fluke Oscilloscopes Items Including:
Fluke Scopemeter Model 125/003S, 40 MHz with Software (FLUKE-125/003S)
Fluke 125 Industrial Scopemeter Oscilloscope
Fluke BP291 Li-ion Battery pack, double capacity, for Fluke 190 Series
Fluke EBC290 External battery charger, LI-ion battery, BP291.
More Details
Cell Culture Supplies
Buy Cell Culture Supplies Items Including:
Thermo Scientific Nunc Nunclon™ cell culture dish, vented, 21 cm2,
Thermo Scientific Nunc Nunclon™ cell culture dish, vented with grid.
More Details
Electric Thermometers
Buy Electric Thermometers Items Including:
Digital multimeter/infrared thermometer with laser sighting Dot/Circle (HHM290)
Fluke 62/T+Pro/1AC II, Electrical/HVAC Kit, 62 Mini IR Thermometer, T+ (FLUKE-62/T+PRO/1AC)
Extech EX845, CAT IV 1000 A Clamp Meter with IR Thermometer (YO-20036-54).
More Details
70618
results
<<<Previous
36
37
38
39
40
41
42
43
44
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
10
20
25
26
27
28
29
30
31
32
33
34
35
36
37
38
39
40
41
42
43
44
45
46
47
48
49
50
51
52
53
54
60
70
80
90
100
110
120
130
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Kleenpak Fittings to Electric Thermometers
>
31125329-s-toc_2 31125329stoc2