Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 5

Scienceware Hydrometer Racks   to   Cole-Parmer Nylon Manifolds

Image
  Product Detail
More Detail

Scienceware Hydrometer Racks

Buy Scienceware Hydrometer Racks Items Including:
  • Hydrometer Rack for Hydrometers Greater Than 280 mm Long (18988-0000).

Lab Coat Safety

Buy Lab Coat Safety Items Including:
  • Tyvek Lab Coat, Large
  • META 100% Cotton Lab Coat, Men's X-Large (48)
  • META 100% Cotton Lab Coat, Women's Small (8)
  • Flame-Resistant Lab Coat, Light Blue, small
  • Flame-Resistant Lab Coat, Light Blue, medium

Microtube Tough Spots

Buy Microtube Tough Spots Items Including:
  • Small Tough-Spots™ label, white
  • Small Tough-Spots™ label, yellow
  • Small Tough-Spots™ label, green
  • Small Tough-Spots™ label, blue
  • Large Tough-Spots™ label, white

Handle Iron

Buy Handle Iron Items Including:
  • Cast iron float and thermostatic steam trap (in-line horizontal config (FTI-15 1/2" 66664 )
  • Cast iron float and thermostatic steam trap (in-line horizontal config (FTI-125 3/4" 66672 ).

Burettes and Pipettes

Buy Burettes and Pipettes Items Including:
  • Burette Brush,36", 12/pk
  • Pipette Brush, 17" X 1/8"-3/4", 12/pk
  • Pipette Brush, 17" X 3/16"-5/16", 12/pk.

Tri Clamp Sanitary Fittings

Buy Tri Clamp Sanitary Fittings Items Including:
  • Silicone Gasket, 10 MESH, Size 1-1/2" Tri-Clamp (YO-30523-06)
  • Silicone Gasket, 20 MESH, Size 1/2" Tri-Clamp (YO-30523-20)
  • Silicone Gasket, 20 MESH, Size 1-1/2" Tri-Clamp (YO-30523-26).

Hplc Liquid Chromatography

Buy Hplc Liquid Chromatography Items Including:
  • Cole-Parmer HPLC Column Heater, Compact 90–260 VAC, 50/60 Hz
  • Cole-Parmer HPLC Column Chiller/ Heater (Multi) 90–260 VAC, 50/60 Hz
  • HPLC Column Chiller/ Heater Programmable Oven, 90–260 VAC, 50/60 Hz
  • 1 L HPLC Reservoir, Conical bottom (YO-02017-05).

Static Mixer Sizing

Buy Static Mixer Sizing Items Including:
  • Sanitary Mixer, 1/2" Tri-clamp size, 6 elements (YO-04668-28)
  • Sanitary Mixer, 1/2" Tri-clamp size, 12 elements (YO-04668-30)
  • PVC Mixer with Injection Port, 1-1/2" Line size (YO-04668-62).

Glassware Washers

Buy Glassware Washers Items Including:
  • Undercounter Glassware Washer 115V (YO-98950-01)
  • Freestanding Glassware Washer, 115V (YO-98950-11)
  • Heated Dry Cycle for Cole-Parmer® Glassware Washers (YO-98950-40)
  • FlaskScrubber Undercounter Hi-Temperature Glassware Washer, 230V 50/60 (4420321).

200 Ml Beaker

Buy 200 Ml Beaker Items Including:
  • Pyrex Brand 1060 Berzelius Tall Form Beaker, 200 mL, 12/pk (1060-200).

1000 Ml Beaker

Buy 1000 Ml Beaker Items Including:
  • Pyrex Brand 1000 Griffin Low-Form Beaker, 1000 mL, 24/cs (1000-1L)
  • Polypropylene graduated low-form beaker with handle, 1000 mL
  • Azlon polypropylene "square ratio" beaker, 1000 mL
  • Polycarbonate graduated beaker with handle, 1000 mL (3216).

Siphon Hand

Buy Siphon Hand Items Including:
  • Hand-operated chemical siphon/drum pump, 26 strokes/gallon
  • Hand-operated siphon pump, 5 GPM, Polyethylene (4005(LSS-1345))
  • Hand-operated siphon/lift pail pump, 32 strokes/gallon (9A)
  • Hand-operated siphon/lift drum pump, 16 strokes/gallon (MP1848S).

High Density Polyethylene Bottles

Buy High Density Polyethylene Bottles Items Including:
  • Thermo Scientific Nalgene heavy-duty high-density polyethylene bottle, (2125-1000)
  • Thermo Scientific Nalgene heavy-duty high-density polyethylene bottle, (2125-2000)
  • Thermo Scientific Nalgene heavy-duty high-density polyethylene bottle, (2125-4000).

Pro Balance

Buy Pro Balance Items Including:
  • Acculab Pocke-Pro Balance, 400 g x 0.1 g (PP-401)
  • Acculab Pocket-Pro Balance, 65 g x 0.01 g (PP-62)
  • Ohaus Scout Pro Portable Balance, 200 g, 115 VAC
  • Ohaus Scout Pro Portable Balance, 200 g, 230 VAC
  • Ohaus Scout Pro Portable Balance, 600 g, 115 VAC.

Tube Stoppers

Buy Tube Stoppers Items Including:
  • Conical-Bottom Glass Centrifuge Tubes, 5 mL, Penny-Head Stopper, 12/ca (8084-5)
  • Conical-Bottom Glass Centrifuge Tubes, 15 mL, Penny-Head Stopper (8084-15)
  • Empty Tubes with cork stoppers
  • Set of 144
  • Cork stoppers for sample tubes 08710-50 (PV-0576).

Touch-N-Tuff Disposable Nitrile Gloves

Buy Touch-N-Tuff Disposable Nitrile Gloves Items Including:
  • Ansell Touch-N-Tuff™ Nitrile Gloves, Powder-free, Small
  • Ansell Touch-N-Tuff™ Nitrile Gloves, Powder-free, Medium
  • Ansell Touch-N-Tuff™ Nitrile Gloves, Powder-free, Large
  • Ansel Touch-N-Tuff™ Nitrile Gloves, Powdered, Extra-large.

Cole-Parmer Leather-Palm Work Gloves

Buy Cole-Parmer Leather-Palm Work Gloves Items Including:
  • Economical Leather-Palm Work Gloves, gauntlet cuffs; one size fits all (7612)
  • Quality Leather-Palm Work Gloves, safety cuffs; size, large; pack of 1.

Disposable Plastic Pipettes

Buy Disposable Plastic Pipettes Items Including:
  • Transfer Pipette, 1.5ml, Graduated, 500/pk (YO-06226-37)
  • Transfer Pipette, 3.9ml, Graduated, 500/pk (YO-06226-41)
  • Transfer Pipette, 5.8ml, Graduated, Large Bulb, 500/pk (YO-06226-61)
  • Transfer Pipette 7.5ml Graduated Large Bulb 500/pk (YO-06226-63).

Thermal Imaging Software

Buy Thermal Imaging Software Items Including:
  • FLIR E40 Thermal Imaging Camera BenchKit w/ Software
  • FLIR T420 Thermal Imaging Camera BenchKit w/ Software
  • FFLIR T420 Thermal Imaging Camera BenchKit w/ Software and Close Up Le
  • FLIR Tools+ Analysis and Report Generation Software for 2 users (T127.

Cimarec Stirrer

Buy Cimarec Stirrer Items Including:
  • Thermo Scientific Cimarec ceramic stirrer, 4x4 120v
  • Thermo Scientific Cimarec ceramic stirrer, 10x10 120v
  • Thermo Scientific Cimarec i Telesystem 6 position Stirrer with Telemod
  • Thermo Scientific Cimarec Immersible Six-Position Stirrer, 7 Watt, 230.

Safety Waste Cans

Buy Safety Waste Cans Items Including:
  • Thermo Scientific Nalgene Fluorinated HDPE Safety Waste System
  • Biohazard Waste Can, 10 gallon
  • Biohazard Waste Can, 14 gallon
  • Thermo Scientific Nalgene Biohazard Waste Container, 1.5 gal.
  • Thermo Scientific Nalgene Biohazard Waste Container, 5 gal..

Nalgene Tissue Culture

Buy Nalgene Tissue Culture Items Including:
  • Flexible Top Works™ Closures for Thermo Scientific Nalgene Bottles (2135-3803)
  • Flexible Top Works™ Closures for Thermo Scientific Nalgene Carboys, (2135-8300)
  • Flexible Top Works™ Closures for Thermo Scientific Nalgene Carboys, (2135-8302).

Cole-Parmer Analog Micrometer

Buy Cole-Parmer Analog Micrometer Items Including:
  • 52-229-201:1in Economy Micrometer (52-229-201).

Cole-Parmer Flexible Impeller Pumps

Buy Cole-Parmer Flexible Impeller Pumps Items Including:
  • Flexible impeller pump head, epoxy body/neoprene impeller, 10 GPM, 30.

Tube 16

Buy Tube 16 Items Including:
  • Thermo Scientific Nalgene Oak Ridge round-bottom tubes; 16-mL; PPCO; 1
  • Thermo Scientific Nalgene Oak Ridge round-bottom tubes; 16-mL; PC; 10/
  • Black Rubber Vacuum Tubing, 3/4" ID x 1 1/2" OD, 25ft
  • (YO-96143-16).

Cole-Parmer Nylon Barbed Cross Connectors

Buy Cole-Parmer Nylon Barbed Cross Connectors Items Including:
  • Barbed Fittings; Cross Connector, Nylon, 5 mm Tubing ID, 50/pk (YO-31418-11).

High Temperature Sensors

Buy High Temperature Sensors Items Including:
  • Fluke 68 Infrared Thermometer, Advanced high-temperature model (FLUKE-68)
  • Fluke 568 Infrared Thermometer, Advanced High-Temperature Model (FLUKE-568)
  • High Performance battery-powered turbine flowmeter, 1/2" NPT(M), 0.3 t (HB110-375-S10).

Cole-Parmer Acetal Check Valves

Buy Cole-Parmer Acetal Check Valves Items Including:
  • Acetal Check Valve, 1/4" NPT(F) inlet/1/4" NPT(F) outlet (0354300).

Temperature Labels

Buy Temperature Labels Items Including:
  • Temperature Labels, Refrigerator (YO-90308-80)
  • Temperature Labels, Freezer (YO-90308-85)
  • Temperature Labels, Cooler (YO-90308-90)
  • Five-Point temperature indicating labels; temperature points 105-130d (YO-90303-20).

Cole-Parmer Nylon Manifolds

Buy Cole-Parmer Nylon Manifolds Items Including:
  • Nylon manifold; 2 outlets; 1/4" inlet NPT(F) (YO-31522-31)
  • Nylon manifold; 2 outlets; 3/8" inlet NPT(F) (YO-31522-33)
  • Nylon manifold; 2 outlets; 1/2" inlet NPT(F) (YO-31522-35).

70618 results


Scienceware Hydrometer Racks to Cole-Parmer Nylon Manifolds

>