Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 50
Delmhorst Bd-2100
to
Regulator with Gauge
Image
Product Detail
More Detail
Delmhorst Bd-2100
Buy Delmhorst Bd-2100 Items Including:
BD-2100 basic package
BD-2100 EIFS package
BD-2100 flooring package.
More Details
Radiation Safety Sign
Buy Radiation Safety Sign Items Including:
Safety Sign, Caution - Radiation Area, 7" X 10", Adhesive Vinyl
Safety Sign, Caution - Radiation Area, 7" X 10", Aluminum
Safety Sign, Caution - Radiation Area, 10" X 14", Adhesive Vinyl.
More Details
16 Mesh
Buy 16 Mesh Items Including:
Spectra/Mesh Screen Discs, Polypropylene, 47 mm, 105 µm, pack of 10
Sock Screen Gasket, Platinum-cured Silicone, 16-ss Mesh, 0.75 Length, (SSG-PX-S-150/16SS)
Sock Screen Gasket, Platinum-cured Silicone, 16-ss Mesh, 0.75 Length, (SSG-PX-S-200/16SS).
More Details
Wheel Technologies
Buy Wheel Technologies Items Including:
P5251:Mtlx 0.5in1inMINITAP All Metal Flo Sensor (P526-2005)
P526-2010 1.0in Metalex Minitap (P526-2010)
Compact Output Module, PVC, for1 1/2" pipe; 5.5 to 44 GPM (US08466)
Digital Flow Sensor, PVC, for 1/2" pipe; 0.61 to 18 GPM (USO8402).
More Details
Hardware Stainless
Buy Hardware Stainless Items Including:
Stainless steel mounting hardware; mounts one (1) Masterflex L/S 36 St
Stainless steel mounting hardware; mounts two (2) Masterflex L/S 36 St
Stainless steel mounting hardware; for one L/S Easy-Load II pump head..
More Details
Asco Red-Hat Asco Valves
Buy Asco Red-Hat Asco Valves Items Including:
Brass, Two-Way, Pilot-Operated Valves (Normally Closed), 1-1/4" NPT(F) (8210G8,110/50,120/60)
Brass, Two-Way, Pilot-Operated Valves (Normally Closed), 1-1/2" NPT(F) (8210G22,110/50,120/60).
More Details
For Carts
Buy For Carts Items Including:
Stainless Steel Equipment Cart without drawers; 18-3/8" x 16-5/8", 3 s
SS equipment cart without drawers; 18-3/8" x 16-5/8", 3 shelves, 400 l
SS equipment cart with drawers; 18-3/8" x 16-5/8", 3 shelves, 200 lb c (356).
More Details
Compression Fittings
Buy Compression Fittings Items Including:
Inserts for soft tubing for compression fittings, white PP, 5/16" OD x (YO-06391-20)
Compression fittings, male pipe adapter, straight, Black PP, 1/4" OD x (YO-06444-01).
More Details
Thermal Flowmeters
Buy Thermal Flowmeters Items Including:
Thermal Non-Invasive Low-Flow Flowmeters, 50 to 1500 nL/min (YO-32611-00)
Thermal Non-Invasive Low-Flow Flowmeters, 250 to 7000 nL/min (YO-32611-02)
Thermal Non-Invasive Low-Flow Flowmeters, 200 to 1000 µL/min (YO-32611-08).
More Details
Solar Digital Thermometer
Buy Solar Digital Thermometer Items Including:
Solar Power Thermometer (4123CP)
Solar-powered adjustable-angle fahrenheit thermometer;2.5"L (TI.82)
Solar-powered adjustable-angle fahrenheit thermometer;4"L (TI.82)
Solar-powered adjustable-angle fahrenheit thermometer;6"L (TI.82).
More Details
Cleanroom Vinyl Gloves
Buy Cleanroom Vinyl Gloves Items Including:
QRP Vinyl Cleanroom Gloves, small, 100/bag
QRP Vinyl Cleanroom Gloves, medium, 100/bag
QRP Vinyl Cleanroom Gloves, large, 100/bag.
More Details
Kynar High-Purity Plastic Diaphragm Valves
Buy Kynar High-Purity Plastic Diaphragm Valves Items Including:
Valve, Kynar with EPDM diaphragm, 4.0 CV (KDV075)
Valve, Kynar with EPDM diaphragm, 12.3 CV (KDV150).
More Details
Scientific Industries Programmable Stirrers
Buy Scientific Industries Programmable Stirrers Items Including:
MagStir Genie Compact Programmable Stirrer 120 VAC
MagStir Genie Compact Programmable Stirrer 230 VAC
Multimagstir Genie, 4-position, 120 VAC.
More Details
Sump 10"
Buy Sump 10" Items Including:
Double open end high-temperature housing with 304 SS sump, 10" (156017-02)
Double open end hot water housing with glass-reinforced nylon sump and (150015)
Double open end cold water housing with transparent styrene acrylonitr (150436).
More Details
Metal Tanks
Buy Metal Tanks Items Including:
Floorstand for Thermo Scientific Barnstead Metal Classic Stills Storag (H1000).
More Details
Hepa Systems
Buy Hepa Systems Items Including:
Primary HEPA Filter Package with Carbon Module for Air Cleaning System (YO-33703-04)
Replacement Prefilters for HEPA Filter for Air Cleaning Systems, Pack (YO-33703-12)
HEPA Filter for use with Ductless Air Filter System (YO-33703-43).
More Details
Olympic Optical Eyewear and Safety Glasses
Buy Olympic Optical Eyewear and Safety Glasses Items Including:
Olympic Optical magnifying safety glasses, black frame w/clear lens, + (OL30-06C1.5I)
Olympic Optical magnifying safety glasses, black frame w/clear lens, + (OL30-06C2.0I).
More Details
Stainless Carts
Buy Stainless Carts Items Including:
Stainless Steel Equipment Cart without drawers; 18-3/8" x 16-5/8", 3 s
SS equipment cart without drawers; 18-3/8" x 16-5/8", 3 shelves, 400 l
SS equipment cart with drawers; 18-3/8" x 16-5/8", 3 shelves, 200 lb c (356).
More Details
Masterflex Remote Control Connectors
Buy Masterflex Remote Control Connectors Items Including:
DB25 male connector; for creating your own cable
(07523-94)
DB9 male 9-pin connector for use with Masterflex drives.
DB15 male connector; for creating your own cable
(07595-52)
More Details
High Temperature Pressure Switch
Buy High Temperature Pressure Switch Items Including:
Stainless steel liquid flow switch for high inline pressures; 0.5 GPM (YO-32774-42)
Stainless steel liquid flow switch for high inline pressures; 1.0 GPM (YO-32774-44)
UL-approved Liquid Flow Switch for High Inline Pressures; 0.10 GPM (YO-32778-06).
More Details
Ashcroft Pressure Transducer
Buy Ashcroft Pressure Transducer Items Including:
Differential Pressure Transducer, 0.25" Water Column, 4-20 mA output (CXLDP0.25IWC) (YO-68333-20)
Differential Pressure Transducer, 2.0" Water Column, 4-20 mA output
Differential Pressure Transducer, 5.0" Water Column, 4-20 mA output.
More Details
Dwyer Gauges
Buy Dwyer Gauges Items Including:
Dwyer Magnehelic Differential Pressure Gauge, 2002: 0-2.0" WC
Dwyer Minihelic Differential Pressure Gauge, 0-0.5" WC
Dwyer Minihelic Differential Pressure Gauge, 0-1.0" WC
Dwyer Minihelic Differential Pressure Gauge, 0-2.0" WC.
More Details
Cole-Parmer Pressure Filtration
Buy Cole-Parmer Pressure Filtration Items Including:
Cole-Parmer Compact Filtration System, CPVC, 30 psi max pressure
KNF Vacuum/pressure Pump, 11.5 L/min, 218 Torr, 30 PSIG, 115 VAC (N811 KTP-IP20 (115 VAC))
KNF Vacuum/pressure Pump, 11.5 L/min, 218 Torr, 30 PSIG, 115 VAC (N811 KNP-IP20 (115 VAC)).
More Details
Microscopic Illuminators
Buy Microscopic Illuminators Items Including:
Standard microscope stand; illumination source, 6 V, 10 W halogen bulb (PBH STAND)
Standard microscope stand; illumination source, 6 V, 10 W halogen bulb (PBH STAND-220VAC)
Wide-View Dimmable Illuminated Magnifier, 25" reach, 3 diopters, table.
More Details
Biological Oxygen Monitor
Buy Biological Oxygen Monitor Items Including:
YSI 5300A Biological Oxygen Monitor, 115 VAC
YSI 5300A Biological Oxygen Monitor, 230 VAC
Bath assembly, 115 VAC for YSI biological oxygen monitoring system.
More Details
Hazardous Material Safety
Buy Hazardous Material Safety Items Including:
Hazardous Materials Identification Label, 4" X 4"
Vinyl Hazardous Materials Warning Label, 4"w X 6"h
Hazardous Material Identification Label, 3"W X 5"H
Hazardous Material Identification Label, 7/8" X 7/8".
More Details
Cole-Parmer Low-Cost Hazardous-Duty Air-Drive Mixers
Buy Cole-Parmer Low-Cost Hazardous-Duty Air-Drive Mixers Items Including:
Low-cost, gear-drive air motor mixer with steel gears; 50-1200 rpm (YO-04681-10)
Low-cost, gear-drive air motor mixer; 50-1200 rpm (YO-04685-00)
Low-cost, Direct-drive Air Motor Mixer; 200-7,000 RPM (YO-04686-00).
More Details
Weiss Pressure & Vacuum Instruments
Buy Weiss Pressure & Vacuum Instruments Items Including:
Pressure Gauge Vacuum-15 PSI, 1/4" NPT(M); 316 SS Sensor/ 316 SS Socke
Pressure Gauge Vacuum-60 PSI, 1/4" NPT(M); 316 SS Sensor/ 316 SS Socke
Pressure Gauge Vacuum-100 PSI, 1/4" NPT(M); 316 SS Sensor/ 316 SS Sock.
More Details
Stainless Steel Needle Valves
Buy Stainless Steel Needle Valves Items Including:
Stainless Steel, 2-Way, (Normally Closed), 1/4" NPT(F), 5/32" orifice, (8262H220-120/60)
Stainless Steel, 2-Way, (Normally Closed), 1/4" NPT(F), 7/32" orifice, (8262H226-120/60).
More Details
Regulator with Gauge
Buy Regulator with Gauge Items Including:
Vacuum/pressure Diaphragm Pump With Gauges And Regulators, PTFE-coated (107CA18TFEL-246)
Vacuum/pressure diaphragm pump with gauges and regulators, PTFE-coated (107CDC18TFEL-246).
More Details
70618
results
<<<Previous
46
47
48
49
50
51
52
53
54
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
10
20
30
35
36
37
38
39
40
41
42
43
44
45
46
47
48
49
50
51
52
53
54
55
56
57
58
59
60
61
62
63
64
70
80
90
100
110
120
130
140
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Delmhorst Bd-2100 to Regulator with Gauge
>
31125329-s-toc_2 31125329stoc2