Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 500
Extraction Systems
to
Masterflex Silicone (Platinum Cured) Microbore Tubing
Image
Product Detail
More Detail
Extraction Systems
Buy Extraction Systems Items Including:
Extract-All Portable Air Cleaning System with 4' to 6' Long Extractor
Soxhlet extraction glassware systems, 25 mL (YO-99036-00)
Soxhlet extraction glassware systems, 125 mL (YO-99036-10)
Secondary Merv 8 Pleated Filter
Primary Merv 14 Cell Filter.
More Details
Uv Water Systems
Buy Uv Water Systems Items Including:
Thermo Scientifice GenPure x-CAD UV Water Purification System Benchtop
Replacement UV bulb for 99280-00, -05, -10, -15 (#10 (210-100))
Replacement UV Bulb for 2 GPM Combination Filter/Ultraviolet Sterilize (#11 (210-110))
UV Lamp Without Tm (2068).
More Details
Forced Convection
Buy Forced Convection Items Including:
Binder BD Series Natural Convection Incubator 25.7 Cu
Ft
115V (YO-39072-20)
Carbolite Peak Series Forced Convection Incubator 28L
Carbolite Peak Series Forced Convection Incubator 127L
More Details
Stainless Steel Impeller
Buy Stainless Steel Impeller Items Including:
Cole-Parmer Stainless Steel Centrifugal Pump, Open Impeller, 155 GPM o (YO-75506-01)
Cole-Parmer Stainless Steel Centrifugal Pump, Open Impeller, 175 GPM o (YO-75506-03)
Cole-Parmer Stainless Steel Centrifugal Pump, Open Impeller, 230 GPM o.
More Details
50 Tube
Buy 50 Tube Items Including:
Dow Corning Pharma-50 Tubing, 1/8" x ¼", 50-ft pk
Dow Corning Pharma-50 Tubing, 3/16" x 5/16", 50-ft pk
Dow Corning Pharma-50 Tubing, 3/16" x 3/8", 50-ft pk
Dow Corning Pharma-50 Tubing, ¼" x 7/16", 50-ft pk.
More Details
Cole-Parmer Ultimate Acid Eater
Buy Cole-Parmer Ultimate Acid Eater Items Including:
Ultimate Acid Eater; 2.5 Gal.; 2 Packs/case.
More Details
Shelf Trucks
Buy Shelf Trucks Items Including:
Ergonomic Two-Shelf Truck, 24" x 36" (ERGL-8V2-1PYBK)
Ergonomic Two-Shelf Truck, 24" x 48" (ERGL-8V2-2PYBK)
Ergonomic Two-Shelf Truck, 30" x 48" (ERGL-8V2-5PYBK)
MetroMax Utility Cart, 2 Shelf, 36 (MXUC2436G2-25).
More Details
Digital Temperature Probes
Buy Digital Temperature Probes Items Including:
Fluke Integrated Temperature Probe
Temperature Probe, Stainless Steel, for IKA Eurostar Control Digital M
Temperature Probe, Glass-Coated Stainless Steel, for IKA Eurostar Cont
Temperature Probe Extension Cable, for IKA Eurostar Control Digital Mi.
More Details
Gray Cap
Buy Gray Cap Items Including:
HART Smart Connector, PRT (Gray Cap), Spare (YO-93202-59).
More Details
Rack 30
Buy Rack 30 Items Including:
Screen insert for 50-peg modular stainless steel drying racks
Ismatec™ rack-mount minicartridge pump, 4 channels, 20 rpm, 115 VAC
Scienceware test tube rack for 21- to 25-mm tubes
Thermo Scientific Nalgene Unwire™ test tube rack for 30-mm tubes, w.
More Details
Cole-Parmer Hdpe Trays
Buy Cole-Parmer Hdpe Trays Items Including:
Rectangular high-density polyethylene utility tray, 0.75 gallon (P421-01-2510)
Rectangular high-density polyethylene utility tray, 1.1 gallon (P422-01-2510)
Heavy-Duty Bath, HDPE, 7 gal
Heavy-Duty Bath, HDPE, 15 gal.
More Details
Cole-Parmer Straight Barbed Connectors
Buy Cole-Parmer Straight Barbed Connectors Items Including:
Straight1/2" barbed connector, 316 SS, each (YO-30620-06)
Barbed fittings, Straight Connector, Clear PP9 mm ID, 7 mm x 62 mm x 1 (YO-06459-30)
Barbed fittings, Straight Connector, Clear PP,1/16" ID, 1/32", 25/32", (YO-30506-00).
More Details
Etatron Ph/Ion Process Instruments
Buy Etatron Ph/Ion Process Instruments Items Including:
Etatron HDS pH Pump Control System, 10 L/hr, 5 bar max pressure, wall-
Etatron Single-Channel pH/ORP/Chlorine Controller (YO-99672-31)
Etatron Dual-Channel pH/ORP/Chlorine Controller (YO-99672-33).
More Details
Nylon Cleaning Brushes
Buy Nylon Cleaning Brushes Items Including:
Bottle brush, nylon bristles, white
Bottle brush, X-shaped nylon bristles, yellow
Bottle brush, X-shaped nylon bristles, green
Heavy Duty Carboy Brush, 22", Nylon, 12/pk
Round tank brush, X-shaped nylon bristles, yellow, 7-1/2" diameter.
More Details
High Power Microscopes
Buy High Power Microscopes Items Including:
Cole-Parmer High-power Stereozoom Microscope with fluorescent light, 2 (YO-48411-20).
More Details
Mechanical Seal Pumps
Buy Mechanical Seal Pumps Items Including:
Cole-Parmer 304 SS Mechanically Coupled Pump, 40 GPM, 115/230 VAC (YO-70725-00)
Cole-Parmer 304 SS Mechanically Coupled Pump, 40 GPM, 208-230/460 VAC (YO-70725-02)
Cole-Parmer 304 SS Mechanically Coupled Pump, 40 GPM, 208-230/460 VAC (YO-70725-03).
More Details
Gast Pumps
Buy Gast Pumps Items Including:
Vacuum/pressure diaphragm pump with gauges, regulators, and relief val (DAA-V515-ED)
Vacuum/pressure diaphragm pump, single head, 1.1 cfm, 220/240VAC (DOA-P725-BN)
Vacuum/pressure diaphragm pump with gauges, regulators, and relief val (DOA-P704-AA).
More Details
Material Storage Products
Buy Material Storage Products Items Including:
Two-drawer storage unit (9726-A50)
Storage bin; 4-1/8" x 3" x 5-3/8", 24/pack (30-210 YELLOW)
Storage bin; 4-1/8" x 3" x 5-3/8" (30-210 YELLOW)
Storage bin; 4-1/8" x 3" x 7-3/8", 24/pack (30-220 YELLOW).
More Details
Masterflex Peristaltic Pumps
Buy Masterflex Peristaltic Pumps Items Including:
Masterflex Standard pump head for L/S 13 tubing, PC housing/CRS rotor.
Masterflex Standard pump head for L/S 13 tubing, PPS housing/SS rotor.
Masterflex Standard pump head for L/S 14 tubing, PC housing/CRS rotor..
More Details
Histology Equipment
Buy Histology Equipment Items Including:
Cole-Parmer Tissue Flotation Work Center, 8"x8"x2" with slide dryer, 3 (YO-48579-00)
Cole-Parmer Tissue Flotation Work Center, 8"x11"x2" with slide dryer, (YO-48579-04)
Cole-Parmer High-Capacity Slide Dryer 115VAC/60Hz.
More Details
Extech Power Supplies
Buy Extech Power Supplies Items Including:
Extech Analog DC Power Supply, 115/220 VAC
Extech Digital Regulated DC Power Supply, 115/220 VAC
Extech 282275, 600W DC Power Supply, 120 VAC (YO-20036-55)
Extech 282276, 600W DC Power Supply, 230 VAC (YO-20036-56)
Extech DC Power Supply, 30V/1A.
More Details
Based Data Acquisition
Buy Based Data Acquisition Items Including:
Davis Instruments Wireless Download Base Station (used with 8246 & 812
Set 1 consists of: base, 3 NTC radio probes without display, mains uni
Base, radio frequency 2.4 GHz
Thermocouple Based Temperature Recorder with LCD Display (TCTemp2000).
More Details
Stainless Work Tables
Buy Stainless Work Tables Items Including:
Stainless Steel work table, 36"L x 24"W
Stainless Steel work table, 48"L x 24"W
Stainless Steel work table, 72"L x 24"W
Raised countertop-edged work table with an open base; 24"L, 500 lb cap (TVSS-302).
More Details
Oil Viscometer
Buy Oil Viscometer Items Including:
Silicone Standard Viscosity Oil, RT100
Silicone Standard Viscosity Oil, RT250
Silicone Standard Viscosity Oil, RT300
Silicone Standard Viscosity Oil, RT500
Silicone Standard Viscosity Oil, RT1000
More Details
Polyscience Viscometers
Buy Polyscience Viscometers Items Including:
PolyScience Constant-Temperature Viscosity Baths, 27 L, 120 VAC
PolyScience Constant-Temperature Viscosity Bath, 17 L, 240 VAC
PolyScience Constant-Temperature Viscosity Baths, 17 L, 120 VAC.
More Details
Stainless Steel Washers
Buy Stainless Steel Washers Items Including:
Stainless steel basket with top, 8"L x 8"W x 4"H (YO-98950-74)
Kst47 Washer (301514).
More Details
20 Connector
Buy 20 Connector Items Including:
Microbore tubing connector, straight, 20 Gauge
Microbore tubing connector, reducer, 24 to 20 Gauge
Microbore tubing connector, Y, 20 Gauge
Microbore tubing connector, Y reducer, 24 to 20 Gauge
Microbore tubing connector, Y reducer, 20 to 18 Gauge.
More Details
Oakton General Accessories and Replacement Items
Buy Oakton General Accessories and Replacement Items Items Including:
Boot with built in stand for Oakton temp meters (YO-35427-80)
Hands-free kit for Oakton temp meters (YO-35427-85)
External antenna with magnet for Oakton Wireless Datalogger system
Us (YO-35710-30).
More Details
Cole-Parmer Temperature Transmitters
Buy Cole-Parmer Temperature Transmitters Items Including:
Cole-Parmer 300 pH/ORP/Temperature 1/8-DIN Transmitter
Cole-Parmer Toroidal Conductivity transmitter, CPVC, 0 to 10 mS
Cole-Parmer Toroidal Conductivity transmitter, CPVC, 0 to 100 mS
Cole-Parmer Toroidal Conductivity transmitter, PVDF, 0 to 10 mS.
More Details
Masterflex Silicone (Platinum Cured) Microbore Tubing
Buy Masterflex Silicone (Platinum Cured) Microbore Tubing Items Including:
Masterflex Tygon platinum cured silicone extension tubing, 0.64mm ID,
Microbore two-stop tube sets, silicone (platinum-cured); 0.89 mm ID
P (YO-06421-26)
Microbore two-stop tube sets, silicone (platinum-cured); 1.42 mm ID
P (YO-06421-34).
More Details
70618
results
<<<Previous
496
497
498
499
500
501
502
503
504
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
100
200
300
400
410
420
430
440
450
460
470
480
485
486
487
488
489
490
491
492
493
494
495
496
497
498
499
500
501
502
503
504
505
506
507
508
509
510
511
512
513
514
520
530
540
550
560
570
580
590
600
700
800
900
1,000
1,100
1,200
1,300
1,400
2,000
2,354
Extraction Systems to Masterflex Silicone (Platinum Cured) Microbore Tubing
>
31125329-s-toc_2 31125329stoc2