Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 60

Centrifugation Equipment   to   Bel-Art Microbiology

Image
  Product Detail
More Detail

Centrifugation Equipment

Buy Centrifugation Equipment Items Including:
  • Fixed-angle Rotors for 17304-series centrifuges; 12 x 15-mL (YO-17305-60)
  • Rotor, 6-place for 12-mL centrifuge tubes - for use with 17414-series (C880-01)
  • Rotor, 24-place 0.5- to 2-mL microtubes for 17414-series centrifuges (C880-03).

Cole-Parmer Microbore Ptfe Tubing

Buy Cole-Parmer Microbore Ptfe Tubing Items Including:
  • Microbore PTFE Tubing Assortment Pack
  • Ultramicrobore PTFE Tubing, 0.002"ID 0.014"OD, 100 ft/pack
  • Ultramicrobore PTFE Tubing, 0.004"ID 0.016"OD, 100 ft/pack
  • Ultramicrobore PTFE Tubing, 0.006"ID 0.016"OD, 100 ft/pack.

Cole-Parmer Carboy Brushes

Buy Cole-Parmer Carboy Brushes Items Including:
  • Carboy Brush, 22", Nylon, 12/pk
  • Carboy Brush, 34", Nylon, 12/pk
  • Heavy Duty Carboy Brush, 22", Nylon, 12/pk
  • Heavy Duty Carboy Brush, 34", Nylon, 12/pk
  • Carboy Brush, 22", Nylon; Each

Gallon Storage Tank

Buy Gallon Storage Tank Items Including:
  • Oilless Air Compressor with 20 Gallon Storage Tank, 3/4 hp, 115 VAC/60 (5HCD-11T-M550NGX)
  • Oilless Air Compressor with 20 Gallon Storage Tank, 3/4 hp, 230 VAC/60 (5HCD-11TA-M550X)
  • Professional series air compressor, 7.1 cfm, 20 gallon portable tank,.

Temperature Transmitter Rtd

Buy Temperature Transmitter Rtd Items Including:
  • Economical push-button temperature transmitter, thermistor (SEM203TH-1)
  • Twin Temp 3" Back Connect Face, 4" Stem, RTD Transmitter (30040D206G4R1AR)
  • Twin Temp 3" Back Connect Face, 9" Stem, RTD Transmitter 0/250F (30090D206G4R1AR).

Laboratory Chairs and Stools

Buy Laboratory Chairs and Stools Items Including:
  • Sit-stand stool, 17"-22" height, black reinforced plastic base (3505).

Tyvek Critical Environment

Buy Tyvek Critical Environment Items Including:
  • Tyvek cleanroom boot covers; medium, 100 pairs/case
  • Tyvek cleanroom frock; medium, 30/case
  • Tyvek cleanroom frock; LG, 30/case.

Hirschmann Liquid Handling

Buy Hirschmann Liquid Handling Items Including:
  • Solarus Digital Burette 20 mL (9392020)
  • Solarus Digital Burette 50 mL (9392050)
  • Solarus Interface Connection Cable (9564005).

Fluke Pro

Buy Fluke Pro Items Including:
  • Fluke 62/T+Pro/1AC II, Electrical/HVAC Kit, 62 Mini IR Thermometer, T+ (FLUKE-62/T+PRO/1AC)
  • Fluke 1760 TR Basic, Power Quality Recorder, with Trans, no V or I Pro
  • Intellitone Pro 200 Toner
  • Fluke 561 Infrared Thermometer, HVAC Pro model (FLUKE-561).

High Temperature Pressure Transducer

Buy High Temperature Pressure Transducer Items Including:
  • AccuSense™ Pressure Transducer, 30-0" HG
  • AccuSense™ Pressure Transducer, 0-100 PSIG
  • AccuSense™ Pressure Transducer, 0-100 PSIA
  • AccuSense™ Pressure Transducer, 0-150 PSIG
  • AccuSense™ Pressure Transducer, 0-250 PSIG.

Combination Showers

Buy Combination Showers Items Including:
  • Bradley Combination Showers-Shower/eye/facewash station
  • Guardian Equipment Combination Showers, hand operated, ABS plastic
  • Haws Axion MSR™ Combination Shower with ABS plastic Eyewash
  • Haws Axion MSR™ Combination Shower with Stainless Steel Eyewash.

Iris Scissors

Buy Iris Scissors Items Including:
  • Iris scissors, Dissecting-grade, Curved (YO-10918-10)
  • Iris scissors, Dissecting-grade, Straight (YO-10918-12)
  • Iris scissors, Precision-grade, Straight.

Knf Vacuum Pumps with Explosion-Proof Motors

Buy Knf Vacuum Pumps with Explosion-Proof Motors Items Including:
  • Pump, Vacuum Pump with Explosion Proof Motor, Vacuum and Pressure, Alu (YO-07057-84)
  • Pump, Vacuum Pump with Explosion Proof Motor, Vacuum only, Aluminum he (YO-07057-85)
  • Pump, Vacuum Pump with Explosion Proof Motor, Vacuum only, Solid PTFE.

Hach Kits

Buy Hach Kits Items Including:
  • Hach Free Chlorine (USEPA) Test Kit (21055-69)
  • Hach waterproof ISFET pH meter kit
  • Turbidity Standards Kit for Hach 2100 N/AN Meters, 0, 20, 200, 1000, a (YO-08391-24)
  • Turbidity Standards Kit for Hach 2100P meters, 0, 20, 100, 800 NTU (YO-08391-60).

Water Samplers

Buy Water Samplers Items Including:
  • Global Water WS700 Peristaltic Water Sampler (YO-99243-70)
  • Global Water WS705 Improved Composite/Discrete Water Sampler (YO-99243-75)
  • Water-Level Logger 3 Foot Range, 3 Foot Cable (WL16U-003-003)
  • Replacement bottle/cap for open water grab sampler.

Photohelic Gauge

Buy Photohelic Gauge Items Including:
  • Photohelic Pressure Switch/Gauge, 0.05-0.20" WC, 1/8" NPT(F)
  • Photohelic Pressure Switch/Gauge, 0-1" WC, 1/8" NPT(F)
  • A3003:Swtch/Gauge 0/3.0inWC Photohelic Pre Switch/Gauge
  • A3100:Swtch/Gauge 0/100inWC Photohelic Pre Switch/Gauge.

Ptfe Double Open End Filter Cartridges

Buy Ptfe Double Open End Filter Cartridges Items Including:
  • Double open end PTFE filter cartridge; 0.1 µm (MFE911AAE, 9.824 in LENGTH).

50 Foot Hose

Buy 50 Foot Hose Items Including:
  • Allegro One Man Full Face Piece Supplied Air System with 50 foot Hose
  • Allegro One Man Tyvek Hood Supplied Air System with 50 foot Hose
  • Tygoprene Tubing 1/16 X 3/16, 50 Foot Roll (YO-95870-00)
  • Tygoprene Tubing 1/8 X 1/4, 50 Foot Roll (YO-95870-02).

12 Dc Power Supply

Buy 12 Dc Power Supply Items Including:
  • Tektronix PWS4721 Programmable DC Power Supply, 72 V, 1.2 A
  • Fixed DC power supply; 12 VDC, 150 mA output (YO-26900-05).

Panel Mount Needle Valve

Buy Panel Mount Needle Valve Items Including:
  • Chemically Inert Panel-Mount Needle Valve, 3/8" tube OD (MV-13-163)
  • Chemically Inert Panel-Mount Needle Valve, 1/2" tube OD (MV-13-166)
  • Chemically Inert Panel-Mount Needle Valve, 1/8" tube OD (MV-13-100).

Stainless Steel Relief Valves

Buy Stainless Steel Relief Valves Items Including:
  • Plas Labs Glove Box Pressure Relief Valve, 1/8" NPT Stainless Steel
  • Vent Relief Valve, 100psi (301825)
  • Unloader (Pressure Relief) Valve, Stainless Steel, 75 to 500 PSI Stain.

Radiation Alert Safety Products

Buy Radiation Alert Safety Products Items Including:
  • Radiation Alert® Radiation monitor (DIGILERT 100)
  • Radiation Alert Radiation Monitor
  • Radiation Detector for Radiography, Energy-Compensated; 0 to 1000 mRad (MC1K)
  • Radiation Detector, Energy-Compensated, 0 to 50 mRad/hr; 0 to 500 mSv/ (MONITOR4-USV).

Scienceware Thermometer and Tubing Adapters

Buy Scienceware Thermometer and Tubing Adapters Items Including:
  • Scienceware thermometer adapter of PTFE, size 14/20
  • Scienceware thermometer adapter of PTFE, size 10/18
  • Scienceware thermometer adapter of PTFE, size 19/22
  • Scienceware tubing adapter of PTFE, size 24/40 (YO-06140-52).

Red Panel

Buy Red Panel Items Including:
  • PAXT0000, Temperature Panel Meter Thermocouples- T, E, J, K, R, S, B,
  • PAXT0010, Temperature Panel Meter Thermocouples- T, E, J, K, R, S, B,
  • Panel Meter With Thermocouple And RTD Inputs; Green Lcd
  • Panel Meter With Milliamp And VDC Inputs; Green Lcd.

Statical Process Control

Buy Statical Process Control Items Including:
  • Desco Statguard Carpet Protector, 1 Gallon
  • Staticide ESD Safety Shield Spray, 8 oz can
  • Desco Shielding Bag, 3" x 5" (YO-26808-20)
  • Desco Shielding Bag, 4" x 6" (YO-26808-22)
  • Desco Shielding Bag, 4" x 30" (YO-26808-24).

Gc Columns

Buy Gc Columns Items Including:
  • Thermo Scientific TRACE TR-FAME GC Column 120-m length, 0.25-mm ID, 0
  • (YO-01855-64)
  • Thermo Scientific TRACE™ TR-1 Column 10-m Length, 0.10-mm ID, 0.10- (YO-01852-00)
  • Thermo Scientific TRACE™ TR-1 Column 50-m Length, 0.15-mm ID, 0.50- (YO-01852-02).

Uv Ultraviolet

Buy Uv Ultraviolet Items Including:
  • High-intensity, long-wave UV lamp; 20-ft lead, 115 VAC/60 Hz (YO-97600-10)
  • Handheld Mini 365 nm UV Lamp (95-0125-05(UVL-4))
  • Handheld Mini 254/365 Switchable UV Lamp (95-0188-02(UVSL-14P))
  • UV bench lamp; 15 watts, 254 nm, 230 VAC/50 Hz (95-0042-09).

Dickson Data Loggers

Buy Dickson Data Loggers Items Including:
  • LCD display temperature data logger with external platinum RTD probe
  • Pressure logger with display, 0 to 100 PSI
  • Datalogger, large display, temperature 2-channel (SM320)
  • Datalogger, large display, humidity and temperature with remote probe (TM325).

Process Control Systems

Buy Process Control Systems Items Including:
  • Etatron pH Pump Control System, 1 L/hr Flow
  • Etatron DLXB pH/ORP Pump Control System, 8 L/hr, 10 bar max pressure, (DLXB-PH-RX/MBB) (YO-56022-51)
  • Etatron DLXB pH/ORP Pump Control System, 15 L/hr, 4 bar max pressure, (DLXB-PH-RX/MBB) (YO-56022-52).

Bel-Art Microbiology

Buy Bel-Art Microbiology Items Including:
  • SMAC agar, 500g (YO-36152-26)
  • CT-supplement, 16 vials (YO-36152-28)
  • CAYE broth supplement (Carbadox), 16 vials (YO-36152-34)
  • OXFORD Listeria selective supplement, 13 vials (YO-36152-52)
  • Palcam Listeria selective supplement, 16 vials (YO-36152-56).



Centrifugation Equipment to Bel-Art Microbiology

>