Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 7
Gas Detector System
to
Digital Video Equipment
Image
Product Detail
More Detail
Gas Detector System
Buy Gas Detector System Items Including:
ToxiRAE II Personal Gas Monitor, H2S
ToxiRAE II Personal Gas Monitor, SO2
ToxiRAE II Personal Gas Monitor, NO2
ToxiRAE II Personal Gas Monitor, CL2
ToxiRAE II Personal Gas Monitor, HCN
More Details
Tools Calipers
Buy Tools Calipers Items Including:
Dial caliper, 0 to 6" (142)
Dial caliper, 0 to 150 mm (144MM)
Digital Stainless Steel Caliper, 0 to 12" with Fractions," or Milimete (14712)
Digital calipers with ceramic capacitive sensor; 0 to 4"/0 to 100 mm (54-100-004-1).
More Details
Pressure Transducer Accuracy
Buy Pressure Transducer Accuracy Items Including:
AccuSense™ Pressure Transducer, 30-0" HG
AccuSense™ Pressure Transducer, 0-100 PSIG
AccuSense™ Pressure Transducer, 0-100 PSIA
AccuSense™ Pressure Transducer, 0-150 PSIG
AccuSense™ Pressure Transducer, 0-250 PSIG.
More Details
Precision Tools
Buy Precision Tools Items Including:
Cordless Precision Engraver
UltraTech 28 Piece Multi-Blade Precision Set with Zipper Case (YO-97100-01)
UltraTech 5 Piece Precision Slotted & Phillips Screwdriver Set (YO-97100-02)
UltraTech 5 Piece Precision Torx Screwdriver Set (YO-97100-03).
More Details
Portable Data Logging
Buy Portable Data Logging Items Including:
TRMS System Multimeter Kit with Logging Software (M240H)
DranTech Pro Meter with voltage leads, small case, spare fuses, manual (M243H)
DranTech Pro Meter with voltage leads, large case, USB interface, Dran (M243I).
More Details
Shaft Blade
Buy Shaft Blade Items Including:
304 SS four-blade propeller for shaft 04370-02, 75 mm propeller dia (YO-04370-11)
304 SS four-blade propeller for shaft 04370-02, 60 mm propeller dia (YO-04370-12)
304 SS dual-blade propeller for shaft 04370-02, 100 mm propeller dia (YO-04370-18).
More Details
Cole-Parmer Brix Refractometers
Buy Cole-Parmer Brix Refractometers Items Including:
Low-cost Brix refractometer with ATC; 0 to 10% Brix
Low-cost Brix refractometer with ATC, 0 to 32% Brix
Low-cost Brix refractometer, 28 to 62% Brix
Low-cost salt refractometer, 0 to 100 ppt salinity
Low-cost salt refractometer, 0 to 28% salinity.
More Details
Converter Rs485
Buy Converter Rs485 Items Including:
Rs232 To Rs485 Converter (YO-68479-85)
RS232 to RS485 Converter (YO-89820-63).
More Details
Nylon Mesh Filter
Buy Nylon Mesh Filter Items Including:
Filter top cap for 5-L sealed tub with nylon mesh filter bag (YO-02658-68)
Spectra/Mesh Screen Sheets, 30 X 30 cm Nylon, 70 µm, 3/pk
Spectra/Mesh Screen Discs, 55 mm, Nylon, 8 µm, 10/pk
Spectra/Mesh Screen Sheets, 30 X 30 cm Nylon, 5 µm, pack of 3.
More Details
Lockout Tagout Tags
Buy Lockout Tagout Tags Items Including:
Tag, Danger Do Not Operate This Tag And Lock(bilingual Spanish), 5 7/8 (TSP101CTP)
Tag, Danger Do Not Operate This Tag And Lock (bilingual Spanish), 5 7/ (TSP101PTP)
Tag, Danger Do Not Operate This Tag And Lock (bilingual Spanish), 5 3/ (TSP101LTP).
More Details
Oil Gear Pumps
Buy Oil Gear Pumps Items Including:
High-vacuum gear pump systems for rotary evaporators, 115 VAC
High-vacuum gear pump systems for rotary evaporators, 230 VAC
High-vacuum gear pump systems for filtration, 115 VAC.
More Details
Current Data Loggers
Buy Current Data Loggers Items Including:
USB Current Data Logger, Process, 32,000+ readings, 1 year battery lif
MicroLite II Current USB Data Logger
SmartReader Plus 3 current/voltage/temperature data logger; 8 channels (SRP-00.-1.5M)
AEMC Simple Logger II 4-Channel MiniFlex, Model ML914.
More Details
Cleanroom Labels
Buy Cleanroom Labels Items Including:
Cleanroom Labels, Removable, 2" x 2".
More Details
Cole-Parmer Constant Temperature Products
Buy Cole-Parmer Constant Temperature Products Items Including:
Cole-Parmer® Polystat® CC1080 SEALED CIRCULATOR 120V (YO-12123-50)
Cole-Parmer® Polystat® CC1080 SEALED CIRCULATOR 230V (YO-12123-55).
More Details
Sludge Samplers
Buy Sludge Samplers Items Including:
Raven Coretaker Polycarbonate Sludge Sampler, 16-ft (B-10104-16-REDESIGNED)
Raven Coretaker Polycarbonate Sludge Sampler, 24-ft (B-10104-24)
Sludge Judge II Replacement Top Section with Rope
Sludge Judge Ultra Replacement Bottom Section with Valve.
More Details
Spectrum Low Pressure Liquid Chromatography (Lplc)
Buy Spectrum Low Pressure Liquid Chromatography (Lplc) Items Including:
Spectrum Chromatography Column for Aqueous Solutions, 9.4 mL Total Vol (YO-34697-00).
More Details
Chemical Laboratory Supplies
Buy Chemical Laboratory Supplies Items Including:
Chemical Resistant Tray - 18" X 14", 1 Ea (3184031537)
Chemical Resistant Pan - 31" X 25", 1 Ea (6240085269)
Chemical Technician's Ready Reference Handbook, Fourth Edition
Chemical bottle pourer, 30-mm cap size (310374-0003).
More Details
Cryogenic Containers
Buy Cryogenic Containers Items Including:
Thermo Scientific Nalgene Cryogenic freezing container for 18 (1.2 to.
More Details
Insulated Storage
Buy Insulated Storage Items Including:
Justrite® Flammable Storage Safety Cabinet, 45 gallons, manual doo (YO-81790-31)
Dry ice storage chest, 3.75 cu ft (YO-03720-00)
Dry ice storage chest, 2.5 cu ft (YO-03721-00)
Megger MIT420-EN : Advanced CAT IV 600V With Result Storage Industrial.
More Details
Thin Layer Chromatography
Buy Thin Layer Chromatography Items Including:
Whatman® Blotting and Chromatography Papers, Grade 3 CHR, 46 x 57 (YO-34100-10)
Whatman® Blotting and Chromatography Papers, Grade 3 MM CHR, 46 x (YO-34100-12)
10 x 20 Soft layer, Silica Gel G, 250 µm particle, 60 Å po (YO-34101-20).
More Details
Fume Hood Controls
Buy Fume Hood Controls Items Including:
Controlled atmosphere anaerobic glove box; 2 port; 110 VAC, 60 Hz (YO-34750-10)
Temperature and Humidity Controlled glove box; 2 port; 110 VAC, 60 Hz (YO-34750-40)
Temperature and Humidity Controlled glove box; 2 port; 230 VAC, 50 Hz (YO-34750-45).
More Details
Psi Gauges
Buy Psi Gauges Items Including:
PM 620 Pressure Module +/-10 inH2O psi, gauge type (PM 620 008G)
PM 620 Pressure Module +/-1 psi, gauge type (PM 620 01G)
PM 620 Pressure Module +/-3 psi, gauge type (PM 620 02G)
PM 620 Pressure Module +/-10 psi, gauge type (PM 620 04G).
More Details
3m Safety
Buy 3m Safety Items Including:
3M Three-Panel Particulate respirator, N95, Box of 20
3M 7000 Series Silicone Respirator, Small
3M 7000 Series Silicone Respirator, Large
3M Virtua Sport™ Safety Eyewear, Clear frame, Clear lens
3M Virtua™ 1.5 Magnifier Safety Eyewear, Clear Lens.
More Details
Digital Calibrators
Buy Digital Calibrators Items Including:
Hd.T-Handle Pocket Thermometer , With Potentiometer For Calibration (RR994)
Rubber boot for digital thermometer; 8204 (8204)
NIST-Traceable Calibration; Multimeters, 5.5-6.5 digit (YO-17101-30)
Cole-Parmer Dual Channel Flashing LED Big-Digit Timer.
More Details
Testing Strips
Buy Testing Strips Items Including:
LaMotte Alkalinity Test Strips, 0 to 180 ppm
LaMotte Free Chlorine Test Strips, 0 to 10 ppm
LaMotte Free and Total Chlorine Test Strips, 0 to 10 ppm
LaMotte Hydrogen Peroxide Test Strips, 0 to50 ppm
LaMotte Sanitizer Strength Chlorine Test Strips.
More Details
Kontes Round Bottom
Buy Kontes Round Bottom Items Including:
100 mL 19/22 Round Bottom Flask (YO-34706-08)
100 mL 24/40 Round Bottom Flask (YO-34706-10)
250 mL 19/22 Round Bottom Flask (YO-34706-12)
250 mL 24/40 Round Bottom Flask (YO-34706-14)
1000 mL 24/40 Round Bottom Flask (YO-34706-18).
More Details
Qorpak Clear Jars
Buy Qorpak Clear Jars Items Including:
Qorpak Precleaned Clear Jar, wide mouth, 45 mL, PTFE-lined cap, case o
Qorpak Precleaned Clear Jar, wide mouth, 60 mL, PTFE-lined cap, case o
Qorpak Precleaned Clear Jar, wide mouth, 120 mL, pulp/vinyl-lined cap,.
More Details
Oven Probe
Buy Oven Probe Items Including:
Type J Alligator clip oven thermocouple probe; 2"L (050897C-J)
Type K Alligator clip oven thermocouple probe; 2"L (050897C-K).
More Details
Spectrophotometer Applications
Buy Spectrophotometer Applications Items Including:
Jenway Visible Spectrophotometer; 115 VAC
Cole-Parmer Visible spectrophotometer; 335 to 1000 nm wavelength range (YO-83055-10)
Cole-Parmer Scanning UV/Visible Spectrophotometer, 115 VAC
Temperature Control System for Spectrophotometers, 15 to 40°C.
More Details
Digital Video Equipment
Buy Digital Video Equipment Items Including:
Digital 1.3-megapixel camera with image-analyzing software (SP10.0416 MOTICAM 1000)
Extech Portable Digital Microscope/Camera
Motic Digital Microscope Camera, Moticam 1, 1/4" CMOS
Motic Digital Microscope Camera, Moticam 1SP, 1/3" CMOS.
More Details
70618
results
<<<Previous
3
4
5
6
7
8
9
10
11
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Gas Detector System to Digital Video Equipment
>
31125329-s-toc_2 31125329stoc2