Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 70
High Pressure Check Valves
to
Corning Tissue and Cell Culture
Image
Product Detail
More Detail
High Pressure Check Valves
Buy High Pressure Check Valves Items Including:
Economical High-Pressure Single-Piston Pump; 8.0 mL/min Max Flow, 230 (B-100-S-2-CE (#1442))
High-pressure piston pump, single-piston, 0.5 to 5.0 mL/min, 1000 psi, (1384)
High-pressure piston pump, single-piston, 0.5 to 5.0 mL/min, 1000 psi, (1385).
More Details
Thermo Scientific Square/Rectangular Carboys and Jerricans
Buy Thermo Scientific Square/Rectangular Carboys and Jerricans Items Including:
Thermo Scientific Nalgene rectangular high-density polyethylene carboy (2211-0020)
Thermo Scientific Nalgene rectangular high-density polyethylene carboy (2211-0050)
Thermo Scientific Nalgene rectangular high-density polyethylene carboy (2320-0020).
More Details
14 Mesh
Buy 14 Mesh Items Including:
Mesh Screen, Nylon, 1.18 mm sieve opening, 14 mesh
Spectra/Mesh Screen Discs, Polypropylene, 90 mm, 350 µm, 10/pk
Spectra/Mesh Screen Discs, 47 mm, Nylon, 200 µm, pack of 10
Sieve Calibration Standard, mesh 14
Sieve Calibration Standard, mesh 70.
More Details
Ice Containers
Buy Ice Containers Items Including:
Self-contained Ice Cube Maker, Air Cooled, 115 Vac, 60hz (CCU0220AF1)
Ice storage chest (YO-03719-00)
Dry ice storage chest, 3.75 cu ft (YO-03720-00)
Dry ice storage chest, 2.5 cu ft (YO-03721-00)
Dry ice storage chest, 1.6 cu ft (YO-03721-02).
More Details
Wall Pockets
Buy Wall Pockets Items Including:
Organizer wall unit, blue
Organizer wall unit, red
Organizer wall unit, yellow
Organizer wall unit, green
Organizer wall unit, brown
More Details
Way Micro
Buy Way Micro Items Including:
Cole-Parmer Two-Way Micro Solenoid Rocker Valve, 1/4-28 UNF
Cole-Parmer Two-Way Micro Solenoid Rocker Valve, Manifold Mount
Cole-Parmer Three-Way Micro Solenoid Rocker Valve, 1/4-28 UNF
Cole-Parmer Three-Way Micro Solenoid Rocker Valve, 1/8 NPT(F).
More Details
Progressing Cavity
Buy Progressing Cavity Items Including:
Moyno progressing cavity pump, phenolic housing and rotor/nitrile stat (20551)
Moyno progressing cavity pump, 316 stainless steel housing and rotor/n (3910000251)
Moyno progressing cavity pump, phenolic housing and rotor/nitrile stat (3910000252).
More Details
Variable Area Meters
Buy Variable Area Meters Items Including:
Flowmeter, 30-300 mL/min (YO-32550-01)
Flowmeter 100 - 1000 mL/min (YO-32550-03)
Flowmeter 200 - 2000 mL/min (YO-32550-05)
Flowmeter 300 - 3000 mL/min (YO-32550-07)
Flowmeter 500 - 5000 mL/min (YO-32550-09)
More Details
Spin Tube
Buy Spin Tube Items Including:
Costar Spin-X Centrifuge Tube Filters, Spinfilter Ca 0.22um 100/cs
Costar Spin-X Centrifuge Tube Filters, Spinfilter Ca 0.45µm 100/cs
Costar Spin-X Centrifuge Tube Filters, Spinfilter Ny 0.22um 200/cs.
More Details
Thermo Scientific Stir Bars
Buy Thermo Scientific Stir Bars Items Including:
Thermo Scientific Komet 50 Stirring Bar
Thermo Scientific Komet 30 Stir Bars 5/pk
Thermo Scientific Komet 15 Stir Bars 5/pk
Thermo Scientific Komet Stir Bar 90 with glide ring
Thermo Scientific Komet Stir Bar 50 with glide ring.
More Details
Polyscience Baths & Circulators
Buy Polyscience Baths & Circulators Items Including:
PolyScience MX 7L Refrigerated Bath Circulator 120V/60Hz
PolyScience MX 7L Refrigerated Bath Circulator 240V/50Hz
PolyScience Standard Digital 15L Heating Bath Circulator 120V/60Hz
PolyScience Standard Digital 15L Heating Bath Circulator 240V/50Hz.
More Details
Coalescing Air Filters
Buy Coalescing Air Filters Items Including:
Mini Coalescing Air Filter, 0.3 micron coalescing filter, Max 49 SCFM,
Compac Coalescing Air Filter, 0.3 micron coalescing filter, Max 73 SCF
Mid Size Coalescing Air Filter, 0.3 micron coalescing filter, Max 156.
More Details
Small Bottles
Buy Small Bottles Items Including:
Wheaton Small Bottle Benchtop Roller Culture System, 120V North Americ
Wheaton Small Bottle Benchtop Roller Culture System, 100V Japan
Wheaton Small Bottle Benchtop Roller Culture System, 230V India
Small Bottle/cylinder Brush, 11-1/2", 12/pk.
More Details
Cole-Parmer Hollow Fiber Filters
Buy Cole-Parmer Hollow Fiber Filters Items Including:
Hollow fiber cartridge filter; 0.1 µm, 10"L
Hollow fiber cartridge filter; 0.1 µm, 20"L
Hollow fiber cartridge filter; 0.2 µm, 20"L.
More Details
Dwyer Pressure Gauges
Buy Dwyer Pressure Gauges Items Including:
Dwyer Magnehelic Differential Pressure Gauge, 2000: 0.05-0.20" WC (2000-00N)
Dwyer Magnehelic Differential Pressure Gauge, 2000: 0.05-0.20" WC (2000-00)
Dwyer Magnehelic Differential Pressure Gauge, 200000AV: 0/.25 inWC.
More Details
Digital Flowmeters
Buy Digital Flowmeters Items Including:
A109GMN025NA1:Flmtr .3/3GPM Digital Flowmeter W/ 1/4inNYLON Turbine Ho (A109GMN025NA1) (YO-05610-01)
A109GMN025NA1:Flmtr .3/3GPM Digital Flowmeter W/ 1/4inNYLON Turbine Ho (A109GMN025NA1) (YO-05610-02).
More Details
Oakton Conductivity Meter
Buy Oakton Conductivity Meter Items Including:
Oakton conductivity solution, 12,880 µS
Oakton® conductivity solution, 1413 µS
Oakton® conductivity solution, 2764 µS
Oakton® conductivity solution, 2070 µS
Oakton® conductivity solution, 80,000 µS
More Details
Carchip Dataloggers
Buy Carchip Dataloggers Items Including:
CARCHIP PRO
Davis Instruments DriveRight 600E OBD version
Davis Instruments Onboard Smartcard Reader for use with 8126OBD
Davis Instruments Smartcards for use with 8126OBD, pack of 10
Davis Instruments Starter Interrupter Kit for use with 8126OBD.
More Details
Vistek Balances & Scales
Buy Vistek Balances & Scales Items Including:
Vistek™ Balance Isolation Platform, 8"W x 10"D
Vistek™ Balance Isolation Platform, 10"W x 15"D.
More Details
24 Power Supply Vac
Buy 24 Power Supply Vac Items Including:
Power supply; 120 VAC, 60 Hz to 24 VDC (104-10-08).
More Details
Push to Connect
Buy Push to Connect Items Including:
John Guest Push-to-Connect Collet Cover, 5/16" Tube OD
Push-to-connect male threaded stem adapter, 1/4" x 1/8" NPTM, 10/pack (YO-34006-66)
Push-to-connect male threaded stem adapter, 1/4" x 1/4" NPTM, 10/pack (YO-34006-67).
More Details
Cole-Parmer Platinum-Cured Silicone Tubing
Buy Cole-Parmer Platinum-Cured Silicone Tubing Items Including:
Platinum-Cured Silicone Tubing, 3/8"ID x 9/16"OD, 25 ft/pack
Platinum-Cured Silicone Tubing, 1/2"ID x 11/16"OD, 25 ft/pack
Platinum-Cured Silicone Tubing, 5/8"ID x 13/16"OD, 25 ft/pack.
More Details
John Guest Push-to-Connect Fittings
Buy John Guest Push-to-Connect Fittings Items Including:
John Guest Push-to-Connect Collet Cover, 5/16" Tube OD
Push-to-connect soft tube supports, 1/2" stem x 3/8" tube OD, 10/pack (YO-34007-39)
Push-to-connect soft tube supports, 5/16" stem x 1/4" tube OD, 10/pack (YO-34007-40).
More Details
Ultraviolet Equipment
Buy Ultraviolet Equipment Items Including:
Long-wave ultraviolet wand; 15 watts, 115 VAC/60 Hz (95013001)
Handheld Mini 254 nm Ultraviolet Lamp (95-0158-04(UVG-4))
Field ultraviolet viewing cabinet (95007202)
Water-resistant and portable ultraviolet lamp rechargeable battery pac (RBP-1270).
More Details
Vane Anemometers
Buy Vane Anemometers Items Including:
Davis Instruments LED Miniature Vane Anemometer
General Tools Economical Vane Anemometer with Case w CFM
Cole-Parmer Vane Thermoanemometer
Cole-Parmer Vane Thermoanemometer/Hygrometer Pen
Cole-Parmer Vane Thermoanemometer/Hygrometer/Dew Point Pen.
More Details
Sealplate Microplates
Buy Sealplate Microplates Items Including:
SealPlate films, 100/pk (YO-13024-00)
SealPlate films, 100/pk, sterile (YO-13024-02)
SealPlate ThinSeal™ films, 100/pk
SealPlate ThinSeal™ films, 100/pk, Sterile
MiniStrip film, 200/pk (YO-13024-04).
More Details
Waterproof Labels
Buy Waterproof Labels Items Including:
Waterproof label, 2"W x 1"H
Waterproof label, 3"W x 1.5"H
Waterproof label, 4"W x 2"H.
More Details
Thin Tubing
Buy Thin Tubing Items Including:
Cole-Parmer PCR Tubes, 0.2 mL, 8-tube strips, thin walled, virgin PP
Cole-Parmer PCR Tubes, individual, 0.2 mL, thin walled, virgin PP
Cole-Parmer PCR Tubes, individual, 0.2 mL, thin walled with dome caps,.
More Details
Cole-Parmer Aprons
Buy Cole-Parmer Aprons Items Including:
PVC/Nylon Aprons, 50"L x 35"W, 12/case
Disposable Polyethylene Aprons, 28"W X 46"L, 100/bag.
More Details
Corning Tissue and Cell Culture
Buy Corning Tissue and Cell Culture Items Including:
Corning cell culture flask, 25 cm2, canted neck, vented cap
Corning cell culture flask, 75 cm2, canted neck, phenolic c
Corning cell culture flask, 75 cm2, canted neck, vented cap
Corning cell culture flask, 150 cm2, canted neck, plug seal.
More Details
70618
results
<<<Previous
66
67
68
69
70
71
72
73
74
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
10
20
30
40
50
55
56
57
58
59
60
61
62
63
64
65
66
67
68
69
70
71
72
73
74
75
76
77
78
79
80
81
82
83
84
90
100
110
120
130
140
150
160
200
300
400
500
600
700
800
900
1,000
2,000
2,354
High Pressure Check Valves to Corning Tissue and Cell Culture
>
31125329-s-toc_2 31125329stoc2