Subtotal =  
Items =  
View Cart
Welcome
 

Product Index Page 700

Amprobe Electrical and Electronics   to   Thermo Scientific Star a Accessories

Image
  Product Detail
More Detail

Amprobe Electrical and Electronics

Buy Amprobe Electrical and Electronics Items Including:
  • ACD-16 PRO: 1000A DMM Data Logging Clamp-On Multimeter (ACD)
  • TRMS Digital Multimeters with PC Connection, Temperature (AM-160)
  • Hot Stick for Tic Tracer (200035-79) (YO-20008-43)
  • Acd-56-Hpq:Power Quality Clamp-On Meter (ACD).

Cyclograph Life Science

Buy Cyclograph Life Science Items Including:
  • Prescraped 2000 micron rotor for CycloGraph Chromatography System (YO-34106-22)
  • Prescraped 4000 micron rotor for CycloGraph Chromatography System (YO-34106-24)
  • Prescraped 6000 micron rotor for CycloGraph Chromatography System (YO-34106-26).

High Pressure Hose Clamps

Buy High Pressure Hose Clamps Items Including:
  • 316 Stainless Steel Tubing, high pressure; 1/16" OD; two 3-ft sections (YO-03300-02).

Excel Life Science

Buy Excel Life Science Items Including:
  • Excel Scientific X-Pierce™ Precut Piercable Sealing Films, 50/pk - (YO-13024-42)
  • ThermalSeal™ Film for Thermal Cycling, Transparent Polyester, for r (YO-13024-76)
  • ThermalSeal™ Film for Thermal Cycling, Transparent Polyester, for r (YO-13024-78).

Teledyne Life Science

Buy Teledyne Life Science Items Including:
  • Foxy R2 Fraction Collector with 2 Racks; each rack holds 36 centrifuge (YO-01465-70)
  • Foxy R2 Fraction Collector with 2 Racks; each rack holds 36 scintillat (YO-01465-80)
  • Foxy R1 Fraction Collector with 1 Rack; rack holds 72 mini vials (YO-01466-50).

Salisbury Tool Kits

Buy Salisbury Tool Kits Items Including:
  • 60 Piece Hot Box Insulated Tool Kit (YO-91100-63).

A&D Weighing Moisture Balances

Buy A&D Weighing Moisture Balances Items Including:
  • A&D Moisture Determination Balance, ML-50, 51 g x 0.005 g, 115 VAC
  • A&D Moisture Determination Balance, MF-50, 51 g x 0.002 g, 115 VAC
  • A&D Moisture Determination Balance, MS-70, 71 g x 0.0001 g, 115 VAC.

Away Life Science

Buy Away Life Science Items Including:
  • RNase AWAY surface decontaminant, 250 ml bottle
  • RNase AWAY Surface Decontaminant, Case of (12) 250 ml bottles
  • RNase AWAY surface decontaminant, 475 ml spray bottle
  • RNase AWAY Surface Decontaminant, Case of (6) 475 ml spray bottles.

Peek Life Science

Buy Peek Life Science Items Including:
  • PEEK high-pressure unions, tees, and crosses; 0.020" bore, union (YO-02008-02)
  • Replacement PEEK ferrule for Upchurch Scientific ™ Fingertight fitt
  • Precolumn filter, 316 SS and PEEK filter, 0.5 µm frit (YO-02011-01).

Vacuum Tubing Pumps & Tubing

Buy Vacuum Tubing Pumps & Tubing Items Including:
  • 316 SS Vacuum tubing, NW 16 flange, 10"
  • Foreline dry ice cold trap, 1/2" OD straight tube connection
  • 90° Elbow Fittings, 100 ISO-K, Aluminum (C100-09-203)
  • Aluminum tee, NW 16 (C105-12-411)
  • Tee Fittings, 100 ISO-K, Aluminum (C100-09-207).

Precision Analytical Balances

Buy Precision Analytical Balances Items Including:
  • Acculab ALC-Series Precision Analytical Balances, 110 g x 0.0001 g (ALC-110.4)
  • UltraClass™ Precision Mass, 1 g (7025-0)
  • UltraClass™ Precision Mass, 20 g (7020-0)
  • UltraClass™ Precision Mass, 50 g (7018-0)
  • UltraClass™ Precision Mass, 200 g (7016-0).

Wheaton Life Science

Buy Wheaton Life Science Items Including:
  • Wheaton TLC Reagent Sprayer Complete Sprayer, 125 mL (YO-34695-10)
  • Wheaton TLC Reagent Sprayer Replacement Head, 125 mL (YO-34695-12)
  • Wheaton TLC Reagent Sprayer Complete Sprayer, 250 mL (YO-34695-16)
  • Wheaton TLC Sample Recovery Tube (YO-34695-26).

Thermo Scientific Biobasic Life Science

Buy Thermo Scientific Biobasic Life Science Items Including:
  • BioBasic 8 Nanobore Column with IntegraFrit - Length 50mm, 75µm ID, (YO-01897-97)
  • BioBasic 8 Nanobore Column with PicoFrit - Length 50mm, 75µm ID, 15 (YO-01897-98)
  • BioBasic 8 Nanobore Column with PicoFrit - Length 50mm, 75µm ID, 15 (YO-01897-99).

Thermo Scientific Nunc Life Science

Buy Thermo Scientific Nunc Life Science Items Including:
  • Thermo Scientific Nunc™ 96-Well Microplates, Immunology, PS, Standa (YO-01928-22)
  • Thermo Scientific Nunc Non-treated 700mL Cell Culture Triple Flask 32/
  • Thermo Scientific Nunc 275mL Non-treated Cell Culture Flask - 100/case.

Centrivap Life Science

Buy Centrivap Life Science Items Including:
  • Labconco CentriVap DNA Centrifugal Concentrator System, 115V (50/60Hz).

Thermo Scientific Super-Nuova Stirrers

Buy Thermo Scientific Super-Nuova Stirrers Items Including:
  • Thermo Scientific Super-Nuova™ ceramic stirring hot plate, 10 x 10 (SP133835Q)
  • Thermo Scientific Super-Nuova™ ceramic stirring hot plate, 10 x 10 (SP133830-33Q)
  • Thermo Scientific Super-Nuova Multiposition Stirring Hot Plate, 4-posi (SP135935Q).

Bto Life Science

Buy Bto Life Science Items Including:
  • Septa BTO, 9.5mm
  • Septa BTO, 11 mm
  • Septa BTO, 9mm
  • Septa BTO, 11.5mm.

Lab Companion Stirrers

Buy Lab Companion Stirrers Items Including:
  • Lab
  • Companion ® Magnetic Stirrer, Ceramic Top, 230V (BDH571055K)
  • Lab
  • Companion® Stirring Hot Plate, Ceramic Top, 240V (BDH571065K)
  • Lab

Abt Electronics Heating Equipment

Buy Abt Electronics Heating Equipment Items Including:
  • Microwave Oven, 1100 Watts, 1.1 Cu
  • Ft (stainless Steel) (JES1142SS)
  • Microwave Oven, 1200 Watts, 2.1 Cu
  • Ft (black) (JE2160BK OR PEB2060BK).

Marathon Life Science

Buy Marathon Life Science Items Including:
  • Septa Marathon™, 9mm (YO-01999-00)
  • Septa Marathon™, 11mm (YO-01999-02)
  • Septa Marathon™, 11.5mm (YO-01999-04)
  • Septa Marathon™, 17mm (YO-01999-06).

Hach Replacement Electrodes

Buy Hach Replacement Electrodes Items Including:
  • pH Electrode, Gel-filled for Portable meter
  • pH Electrode, Double-junction for Portable meter
  • Replacement Sensor for 95941-02; -04; -06.

Cole-Parmer Pumps & Tubing

Buy Cole-Parmer Pumps & Tubing Items Including:
  • Cole-Parmer Drum Pump Tube Set, PP, 39"
  • Cole-Parmer Drum Pump Tube Set, PVDF, 39"
  • Cole-Parmer Drum Pump Tube Set, PVDF, 47"
  • Replacement Tubing Set for Cole-Parmer 73160-XX pumps, Norprene, 0.8,
  • Replacement pump tube, 39", CPVC.

Nalgene Life Science

Buy Nalgene Life Science Items Including:
  • Flexible Top Works™ Closures for Thermo Scientific Nalgene Carboys, (2135-8300)
  • Flexible Top Works™ Closures for Thermo Scientific Nalgene Carboys, (2135-8302)
  • Flexible Top Works™ Closures for Thermo Scientific Nalgene Carboys, (2135-8303).

Eppendorf Life Science

Buy Eppendorf Life Science Items Including:
  • Eppendorf Mastercycler Nexus Thermal Cycler Flat
  • Eppendorf Mastercycler Nexus Thermal Cycler
  • Eppendorf Mastercycler Nexus Thermal Cycler Eco
  • Electroporator, 220 VAC, 50 Hz (YO-36205-05)
  • Electroporation cuvette, 1 mm, 100 µL, sterile (YO-36205-50).

Fluoropolymer Tubing Life Science

Buy Fluoropolymer Tubing Life Science Items Including:
  • Heat-on Insert For 8x16mm Tubes (fluoropolymer Coating) (YO-81228-45)
  • Heat-on Insert For 4x20mm Tubes (fluoropolymer Coating) (YO-81228-50).

Thermo Scientific Hypercarb Life Science

Buy Thermo Scientific Hypercarb Life Science Items Including:
  • Hypercarb Drop-In Guard Cartridge - Length 10mm, 2.1mm ID, 5µm part (YO-01891-42)
  • Hypercarb Drop-In Guard Cartridge - Length 10mm, 4.0mm / 4.6mm ID, 5m (YO-01891-44)
  • Hypercarb Column - Length 100mm, 4.6mm ID, 5µm particle size (YO-01891-95).

Spectra Spectroscopy and Spectrophotometers

Buy Spectra Spectroscopy and Spectrophotometers Items Including:
  • Spectra Library; Nicolet Condensed Phase Academic Sampler, 1000 spectr
  • Spectra Library; Polymer Additives and Plasticizers Library, 1799 spec
  • Spectra Library; Aldrich Solvents Library, 246 spectra
  • Spectra Library; Surfactants Library, 637 spectra.

Hvac/R Life Science

Buy Hvac/R Life Science Items Including:
  • Heating blanket for 3 L vessels (YO-01919-85)
  • Heating blanket for 5 L vessels (YO-01919-86)
  • Heating blanket for 7 L vessels (YO-01919-87)
  • Heating blanket for 15 L vessels (YO-01919-88)
  • Heating blanket for 2 L vessels.

Labtainers Life Science

Buy Labtainers Life Science Items Including:
  • Media Bag, Sterile, 100 mL, 2 Ports (SH30657.12)
  • Media Bag, Sterile, 250 mL, 2 Ports (SH30657.13)
  • Media Bag, Sterile, 500 mL, 2 Ports (SH30657.14)
  • Media Bag, Sterile, 1000 mL, 2 Ports (SH30657.15)
  • Media Bag, Sterile, 2000 mL, 2 Ports (SH30657.16).

Thermo Scientific Star a Accessories

Buy Thermo Scientific Star a Accessories Items Including:
  • Thermo Scientific Orion Electrode Stand For Star Series Benchtop Meter
  • Carrying Case, Star A Meters
  • Armor for Star A Meters
  • Electrode Arm, Star A Series
  • Self-stirring probe for Star-series benchtop meters



Amprobe Electrical and Electronics to Thermo Scientific Star a Accessories

>