Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 8
Sussman Steam Generators
to
Scales Calibration
Image
Product Detail
More Detail
Sussman Steam Generators
Buy Sussman Steam Generators Items Including:
Stainless Steel Steam Generator; 36 Lbs/hr, 208 VAC (SSB12100B3)
Stainless Steel Steam Generator; 36 Lbs/hr, 480 VAC (SSB12100F3)
Electric Steam Generator, with 120 VAC control circuit, 27.0 lb/hr, 20 (MBA9B3).
More Details
Thermo Scientific Hoods
Buy Thermo Scientific Hoods Items Including:
Standing Height Wall Assembly with Fume Hood - 13ft (YO-41524-50)
2 Drawer Wood Lab Table With Epoxy Resin Top 60in W X 30in H X 30in D (YO-41527-20)
1 Drawer Wood Lab Table With Epoxy Resin Top 36in W X 36in H X 30in D (YO-41527-24).
More Details
Fahrenheit Thermometer
Buy Fahrenheit Thermometer Items Including:
Solar-powered adjustable-angle fahrenheit thermometer;2.5"L (TI.82)
Solar-powered adjustable-angle fahrenheit thermometer;4"L (TI.82)
Solar-powered adjustable-angle fahrenheit thermometer;12"L (TI.82).
More Details
Capacitive Switch
Buy Capacitive Switch Items Including:
Flowline Switch-Tek Non-intrusive RF Capacitance Level Switch, PP Brac
Flowline™ SuperGuard™ RF capacitance level switch; sensor materi.
More Details
Cole-Parmer Luer Anti-Siphon Valves
Buy Cole-Parmer Luer Anti-Siphon Valves Items Including:
One-way luer anti-siphon valve, SAN with silicone diaphragm, female-ma (SCV04052).
More Details
Asco Red Hat
Buy Asco Red Hat Items Including:
ASCO Red-Hat steam and hot water valve, 1/2", 300°F
Brass, Two-Way, Pilot-Operated Valves (Normally Closed), 1-1/2" NPT(F) (8210G22,110/50,120/60)
Brass, Two-Way, Pilot-Operated Valves (Normally Closed), 1-1/4" NPT(F) (8210G8,24 VDC).
More Details
Excel Microplates
Buy Excel Microplates Items Including:
Excel Scientific X-Pierce™ Precut Piercable Sealing Films, 100/pk (YO-13024-40)
ThermalSeal™ Films for Thermal Cycling, Polyolefin, 2-mil optically
ThermalSeal A Advanced, non-sterile films, 100/case (YO-13024-58)
SealPlate ThinSeal™ films, 100/pk.
More Details
Cole-Parmer Plastic Buchner Funnels
Buy Cole-Parmer Plastic Buchner Funnels Items Including:
High-density polyethylene Buchner funnel, 55 mm dia (YO-06128-03)
Polypropylene Buchner funnel, 90 mm dia (YO-06128-04)
Polypropylene Buchner funnel, 110 mm dia (YO-06128-05)
Polypropylene Buchner funnel, 150 mm dia (YO-06128-10).
More Details
Cole-Parmer Alkaline Batteries
Buy Cole-Parmer Alkaline Batteries Items Including:
Button cell batteries, 3 V, 6/pack (Eveready # ECR2025) (ECR2025BP)
Batteries, 1.5 V, C, 4/pack (E93VP)
Replacement battery for battery-operated UV lamps, 6 V (1209)
Button cell batteries, 1.5 V, 6/pack (Eveready # 386) (386BP).
More Details
Backlight Display
Buy Backlight Display Items Including:
Extech Stopwatch/Clock with Backlight Display
Timer, Red/Green Display Backlight Display
Digital Manometer with range of 0 to 15 psi
Digital Manometer with range of 0 to 30 psi
Digital Manometer with range of 0 to 100 psi.
More Details
Calibration Hand Pumps
Buy Calibration Hand Pumps Items Including:
Hydraulic Hand Pump, 10,000 PSI
Portable Pressure Calibrator, complete pneumatic pump kit PM305-0010-K
Portable Pressure Calibrator, complete pneumatic pump kit PM305-0020-K
Portable Pressure Calibrator, complete pneumatic pump kit PM305-0040-K.
More Details
General Tools and Instruments Standard Thermohygrometers
Buy General Tools and Instruments Standard Thermohygrometers Items Including:
Digital Psychrometer, RH & Temp (CMM880)
Digital Psychrometer, RH, Temp, & Dew Point (LAM880D).
More Details
Nitrogen Analysis
Buy Nitrogen Analysis Items Including:
RAE Systems gas detection tubes, nitrogen oxides, 1 to 50 ppm, 10/box
CMS Chip, Nitrogen dioxide, 0.5 to 25 ppm
Vapodest Rapid Distillation Systems; Basic distillation system, 115V,
Glassware for Vapodest Distillation Systems, 250mL Kjeldahl flask.
More Details
Amprobe True Rms Power Quality Recorder
Buy Amprobe True Rms Power Quality Recorder Items Including:
Power Quality Recorder, DM-II Plus (DM-II).
More Details
Flow Totalizer
Buy Flow Totalizer Items Including:
Totalizer for Gas Mass Flowmeters and Controllers with Input and Outpu
Flow meter and totalizer mounting kit, no cables
Flow controller and totalizer mounting kit, no cables
Flow meter mounting kit, shielded cable with two 9 pinsD-connectors fo.
More Details
Propellers and Shafts
Buy Propellers and Shafts Items Including:
304 SS four-blade propeller for shaft 04370-02, 75 mm propeller dia (YO-04370-11)
304 SS four-blade propeller for shaft 04370-02, 60 mm propeller dia (YO-04370-12)
304 SS folding propeller for shaft 04370-02, 48 mm propeller dia (open (YO-04370-14).
More Details
Rite in the Rain Notebooks
Buy Rite in the Rain Notebooks Items Including:
Rite in the Rain - all weather copier paper - 8-1/2" x 11"
Rite in the Rain Field Pattern Maxi-Spiral Notebooks
Rite in the Rain Horizontal Line Pattern Maxi-Spiral Notebooks
4-5/8" x 7" spiral notebook - field pattern (353N-6).
More Details
Nylon Hair
Buy Nylon Hair Items Including:
Nylon hair net, 18", white, 100/bg.
More Details
Cole-Parmer Power Supply/Adapters
Buy Cole-Parmer Power Supply/Adapters Items Including:
Power Supply/Adapter (110-00-18T)
Power Supply/Adapter (105-10-08)
Power Supply/Adapter (110-00-08T)
Power supply adapter
Power supply; 120 VAC, 60 Hz to 24 VDC (104-10-08)
More Details
Safety Shower and Eyewash Station
Buy Safety Shower and Eyewash Station Items Including:
Bradley Combination Showers-Shower/eye/facewash station
Safety Sign, Eye Wash Station, 7 X 10, Plastic
Safety Sign, First Aid Station (symbol), 7" X 10", Adhesive Vinyl
Safety Sign, First Aid Station (symbol), 7" X 10", Plastic.
More Details
40 Watt Lamp
Buy 40 Watt Lamp Items Including:
UV bench lamp; 40 watts, 254 nm, 115 VAC/60 Hz (95-0043-07)
UV bench lamp; 40 watts, 254 nm, 230 VAC/50 Hz (95-0043-08)
UV bench lamp; 40 watts, 365 nm, 115 VAC/60 Hz (95-0043-04)
Replacement tube for UV bench lamp; 40 watts, 365 nm (34-0068-02).
More Details
Davis Vantage Pro2 Wireless
Buy Davis Vantage Pro2 Wireless Items Including:
Davis Instruments Vantage Pro2™ Wireless Temperature Station
Davis Instruments Vantage Pro2™ Wireless Integrated Sensor Suite
Davis Instruments Vantage Pro2 Wireless ISS Plus with UV & Solar Radia
UV Sensor for Vanatage Pro2 (wireless) (6490).
More Details
Industrial Thermocouples
Buy Industrial Thermocouples Items Including:
Uniwell Universal Industrial Thermocouple Probes Type J 6"L (1910-J-6-1/2-W2-446)
Uniwell Universal Industrial Thermocouple Probes Type K 6"L (1910-K-6-1/2-W2-446)
Type T Heavy-duty industrial thermocouple probe; 36"L (070897A-T).
More Details
Anti Static Finger Cots
Buy Anti Static Finger Cots Items Including:
QRP Nitrile Anti-Static Fingercots, large, 720/pk
QRP Nitrile Anti-Static Fingercots, extra large, 720/pk.
More Details
Cole-Parmer Low-Cost Transmitters
Buy Cole-Parmer Low-Cost Transmitters Items Including:
Cole-Parmer Low-Cost Gas Mass Flowmeter with acetal Fittings, 0 to 100 (YO-32707-04)
Cole-Parmer Low-Cost Gas Mass Flowmeter with acetal Fittings, 0 to 200 (YO-32707-06).
More Details
Whatman Blotting Papers
Buy Whatman Blotting Papers Items Including:
Whatman 3MM Chr Blotting Paper, 4 x 5.25 in, 100/box
Whatman 3MM Chr Blotting Paper, 11 x 14 cm, 100/box
Whatman 3MM Chr Blotting Paper, 12 x 14 cm, 100/box
Whatman 3MM Chr Blotting Paper, 15 x 17.5 cm, 100/box.
More Details
Ln2 Storage
Buy Ln2 Storage Items Including:
Stainless Steel LN2 Dewar Flask, 1.0 L with Lid (238-111)
Stainless Steel Ln2 Dewar, 1.0l, 7 (238-112)
Stainless Steel Ln2 Dewar, 2.0l, 12-3/8 (238-113)
Stainless Steel Ln2 Dewar, 3.0l, 7-1/2 (238-114)
Stainless Steel Ln2 Dewar, 0.5l, 8 (238-115).
More Details
Talboys Orbital Shakers
Buy Talboys Orbital Shakers Items Including:
Standard Orbital Shaker, 120V.
More Details
Food Quality
Buy Food Quality Items Including:
Masterflex® L/S® variable-speed drive with 10-turn speed contr (77521-57)
Costar small scraper, 18 mm blade x 250 mm handle
MiniStrip film, 200/pk (YO-13024-04)
MiniStrip film, 200/pk, sterile (YO-13024-06)
AccTech Cleaning Pads, TexStat® (CT814).
More Details
Scales Calibration
Buy Scales Calibration Items Including:
Class F Calibration mass, 10kg Cast Iron Painted, Density 7.2 (traceab (12767TR)
Class F Calibration mass, 25kg Cast Iron Painted, Density 7.2 (traceab (12775TR)
Class F Calibration mass, 35kg Cast Iron Painted, Density 7.2 (traceab (12805TR).
More Details
70618
results
<<<Previous
4
5
6
7
8
9
10
11
12
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Sussman Steam Generators to Scales Calibration
>
31125329-s-toc_2 31125329stoc2