Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 80
Cole-Parmer Receiving Pans for Bottom Sieve
to
Stainless Steel Mixers
Image
Product Detail
More Detail
Cole-Parmer Receiving Pans for Bottom Sieve
Buy Cole-Parmer Receiving Pans for Bottom Sieve Items Including:
Receiving Pan for 8" Diameter Stainless Steel Full Height Sieves
Receiving Pan for 8" Diameter Stainless Steel Half Height Sieves
Receiving Pan for 12" Stainless Steel Bottom Sieves, Full Height.
More Details
Amprobe Process Equipment & Instrumentation
Buy Amprobe Process Equipment & Instrumentation Items Including:
Amprobe AM-220 Digital Multimeter, 600 V
Amprobe ACD-51NAV Navigator Clamp Meter, TRMS, 600 A AC
Amprobe ACDC-52NAV Navigator Clamp Meter, TRMS, 600 A AC/DC
Amprobe ACD-53NAV Navigator Clamp Meter, TRMS, 1000 A AC
Amprobe SOLAR-600 Solar Analyzer.
More Details
Fiberglass Trays
Buy Fiberglass Trays Items Including:
Deep Fiberglass Market Pan in Pearl White, 6.5" W x 30" L x 2" D (6302FMT301)
Deep Fiberglass Market Pan in Pearl White, 8.5" W x 30" L x 2" W (8302FMT301)
Red Fiberlglass Tray 10"x13"x3/4; 1/EA (YO-06714-12).
More Details
Astm Glass
Buy Astm Glass Items Including:
Durac Plus ASTM Like 1c (10/001/1)
Durac Plus ASTM Like 1f (10/001/2)
Durac Plus ASTM Like 5c (10/005/1)
Durac Plus ASTM Like 5f (10/005/2)
Durac Plus ASTM Like 6c (10/006/1)
More Details
Rtd Temperature Controller
Buy Rtd Temperature Controller Items Including:
Digi-Sense 1-Zone benchtop dual display temperature control, type RTD (YO-36225-64)
Temperature Controller, RTD, 1/16-DIN, Relay Output, 1 alarm (DC1014CR-101-000-E)
Temperature Controller, RTD, 1/4-DIN, Relay Output, 1 alarms, (DC1044CR-101-000-E).
More Details
Cole-Parmer Air Sampling
Buy Cole-Parmer Air Sampling Items Including:
Casella Tuff™ Air Sampling Pump
Casella Tuff™ Pro Air Sampling Pump
Cole-Parmer Kynar Gas Sampling Bag, 24" x 24", Septum valve (Y3 2424/2001-C)
Cole-Parmer Kynar Gas Sampling Bag, 12" x 12", On/Off and Septum valve (01410-52).
More Details
Sulfide Detector
Buy Sulfide Detector Items Including:
Portable Hydrogen Sulfide Detector, 0 to 50 ppm
GasAlertClip™ Extreme Personal Monitor, Hydrogen sulfide, 2-year op
Replacement Hydrogen Sulfide Sensor, 0 to 100 ppm
GasAlert Extreme Single-Gas Detector, Hydrogen Sulfide, with data logg.
More Details
Digital Radiometer
Buy Digital Radiometer Items Including:
UVX Digital UV Intensity Meter (97-0015-02(UVX))
UV Intensity Meter Sensor, 254 nm (97-0016-01(UVX25))
UV Intensity Meter Sensor, 365 nm (97-0016-02(UVX36))
UV Intensity Meter Sensor, 302 nm (97-0016-04(UVX31)).
More Details
Disposable Nitrile Gloves
Buy Disposable Nitrile Gloves Items Including:
Best Nighthawk™ Black Nitrile Gloves, Small, Box of 50
Best Nighthawk™ Black Nitrile Gloves, Medium, Box of 50
Best Nighthawk™ Black Nitrile Gloves, X-large, Box of 50
Ansell Touch-N-Tuff™ Nitrile Gloves, Powder-free, Small.
More Details
Generator Replacement
Buy Generator Replacement Items Including:
Sight Glass F/steam Generators (99080-1)
Function Generator Lead Set (TLFG).
More Details
Hach Turbidity Benchtop Meters
Buy Hach Turbidity Benchtop Meters Items Including:
Hach 2100N Laboratory Turbidity Meter, EPA compliant, 110 VAC
Hach 2100N Laboratory Turbidity Meter, EPA compliant, 230 VAC
Hach 2100AN Laboratory Turbidity Meter, EPA compliant, 110 VAC.
More Details
Positive Displacement Pump Types
Buy Positive Displacement Pump Types Items Including:
Moderate Flow Positive Displacement Bypass Diaphragm Pump, 1.4 GPM, 11
Moderate Flow Positive Displacement Automatic-Demand Diaphragm Pump, 1 (8020-513-236)
Moderate Flow Positive Displacement Automatic-Demand Diaphragm Pump, 1 (8000-543-236).
More Details
Nema 4 Cabinets
Buy Nema 4 Cabinets Items Including:
Diecast aluminum enclosure, type AN-1303 (AN-1303)
Diecast aluminum enclosure, type AN-1304 (AN-1304)
Diecast aluminum enclosure, type AN-1305 (AN-1305)
Diecast aluminum enclosure, type AN-1306 (AN-1306).
More Details
Viton Ball Valves
Buy Viton Ball Valves Items Including:
Thermoplastic True Union Ball Valve, CPVC with Viton seals, PTFE seats (V07104N)
Thermoplastic True Union Ball Valve, CPVC with Viton seals, PTFE seats (V08104N)
Thermoplastic True Union Ball Valve, CPVC with Viton seals, PTFE seats (V10104N).
More Details
Smart Gasket 2-Port Clamp
Buy Smart Gasket 2-Port Clamp Items Including:
Smart Gasket , 2" Tri-Clamp, 2 Port, Silicone, Each
Smart Gasket , 2" Tri-Clamp, 1 Port, Silicone, Each
Smart Gasket , 2" Tri-Clamp, 3 Port, Silicone, Each
Smart Gasket , 2" Tri-Clamp, 4 Port, Silicone, Each.
More Details
Diaphragm Dosing Pumps
Buy Diaphragm Dosing Pumps Items Including:
Pump, Metering with 5 control methods, 0.66gph, PVDF, Fkm Front Contro (96470133).
More Details
Ika Lab Mixers
Buy Ika Lab Mixers Items Including:
IKA® Lab Egg Compact Mixer, Blue
Three-stage high shear dispersing mixing head for 04300-01 Magic Lab ( (R078352)
IKA Eurostar 20 High-Speed Digital Mixer, 150 to 6000 rpm, 115 VAC
IKA Eurostar 20 High-Speed Digital Mixer, 150 to 6000 rpm, 230 VAC.
More Details
Conical Test Tube
Buy Conical Test Tube Items Including:
Thermo Scientific Nunc Conical Centrifuge Tubes 500/CS, RACKED 15mL
Thermo Scientific Nunc Conical Centrifuge Tubes 300/CS, RACKED 50mL
Conical-Bottom Glass Centrifuge Tubes, 100 mL, for Oil Testing, 12/cas (8200-100).
More Details
Beta Calibration
Buy Beta Calibration Items Including:
BG301-300 : Calibrator, 300psi Betagauge 301 Pressure Calibrator Inclu
Calbrator Kit, 30psi Betagauge 301 Pressure Calibrator Kit
Calibrator Kit, 5psi Betagauge 301 Pressure Calibrator Kit
Calibrator Kit, 1psi Betagauge 301 Pressure Calibrator Kit.
More Details
Alcohol Lamp Burner
Buy Alcohol Lamp Burner Items Including:
Alcohol lamp (19691)
Two-way lamp stand (19693).
More Details
Mixing Heads
Buy Mixing Heads Items Including:
Three-stage high shear dispersing mixing head for 04300-01 Magic Lab ( (R078352)
Powder-liquid dispersing mixing head for 04300-01 Magic Lab (sold sepa (R075333)
Suction-feed powder-liquid dispersing mixing head for 04300-01 Magic L (R075262).
More Details
Cole-Parmer Moderate-Flow Flexible Impeller Pumps
Buy Cole-Parmer Moderate-Flow Flexible Impeller Pumps Items Including:
Self-priming flexible impeller pump, bronze/neoprene, 5.8 GPM, 5.8 psi (18610-0271)
Pump, Flexible Impeller Pump, 20 GPM or 30 PSI, Neoprene Impeller, 1-1
Pump, Flexible Impeller Pump, 8 GPM or 28 PSI, Nitrile Impeller, 3/4 H.
More Details
Plunger Can
Buy Plunger Can Items Including:
Plunger Can, HDPE with SS Fittings, 1 quart
Plunger Can, HDPE with SS Fittings, 4 quart
Plain Plunger; 2 mL X 230 mm (011.202)
Plain Plunger; 5 mL X 235 mm (011.205)
Plain Plunger; 10 mL X 270 mm (011.210)
More Details
Pcr Tube
Buy Pcr Tube Items Including:
Cole-Parmer PCR Tubes, 0.2 mL, 8-tube strips, thin walled, virgin PP
PCR tubes, polypropylene, 0.2 ml, thin wall with flat caps, 1000/case
Cole-Parmer StableTemp Single Block for PCR, 64 single PCR tubes
Cole-Parmer PCR Micro Reaction Tube, 0.5 mL.
More Details
Cole-Parmer Paired Glass and Quartz Cuvettes
Buy Cole-Parmer Paired Glass and Quartz Cuvettes Items Including:
Semimicro cuvettes with frosted thick wall; cover top; glass; 1.4 mL c
Semimicro cuvettes with frosted thick wall; cover top; quartz; 1.4 mL
Micro cuvettes with frosted thick wall; cover top; glass; 0.7 mL capac.
More Details
Ergonomic Microscopes
Buy Ergonomic Microscopes Items Including:
Microscope arm rests (MSM-N-BL)
Microscope arm rests, cleanroom (MSM-CR).
More Details
Extech Datalogging Light Meters
Buy Extech Datalogging Light Meters Items Including:
Extech Heavy-Duty Datalogging Light Meter, Model HD450.
More Details
High Temperature Vacuum
Buy High Temperature Vacuum Items Including:
Electronic-controlled high-temperature vacuum oven, 0.67 cu ft, 120 VA (YO-05017-00)
Microprocessor-controlled high-temperature vacuum oven, 1.50 cu ft, 12 (YO-05017-10)
Marsh Bellofram Type 70 High- Flow Regulators, 1/4" NPT(F), 0 to 2 psi.
More Details
Safety Bottles
Buy Safety Bottles Items Including:
Wheaton Safety bottles with PTFE-lined caps; 16 oz, clear
Wheaton Safety bottles with PTFE-lined caps; 16 oz, amber
Wheaton Safety bottles with PTFE-lined caps; 1 gal., amber
Scienceware Plum Sterile Saline (500-mL bottle)/pH Neutralizing (200-m.
More Details
Stainless Steel Mixers
Buy Stainless Steel Mixers Items Including:
Pipe Mixer, 316 stainless steel, 1/2" NPT(M), 12 elements (YO-04669-18)
Pipe Mixer, 316 stainless steel, 3/4" NPT(M), 12 elements (YO-04669-20)
Pipe Mixer, 316 stainless steel, 1-1/2" NPT(M), 6 elements (YO-04669-23).
More Details
70618
results
<<<Previous
76
77
78
79
80
81
82
83
84
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
10
20
30
40
50
60
65
66
67
68
69
70
71
72
73
74
75
76
77
78
79
80
81
82
83
84
85
86
87
88
89
90
91
92
93
94
100
110
120
130
140
150
160
170
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Cole-Parmer Receiving Pans for Bottom Sieve to Stainless Steel Mixers
>
31125329-s-toc_2 31125329stoc2