Shopping Cart
Subtotal
=
Items
=
View Cart
Live Chat
Welcome
Search
Shop All Products
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Product Index Page 9
Tubing Aluminum
to
Refill Kit
Image
Product Detail
More Detail
Tubing Aluminum
Buy Tubing Aluminum Items Including:
Aluminum Powder Pillow Packs, 10 mL, 100 Tests (RS5000)
Cole-Parmer Aluminum Block, for 40 x 0.5-mL Microcentrifuge Tubes
Cole-Parmer Aluminum Block, for 30 x 1.5-mL Microcentrifuge Tubes
Cole-Parmer Aluminum Blocks, for 12 x 15/16mm ID Test Tubes.
More Details
Brass Frame
Buy Brass Frame Items Including:
Cole-Parmer Testing Sieve, Brass Frame/SS Wire, 8" OD, full height, No (YO-59940-01)
Cole-Parmer Testing Sieve, Brass Frame/SS Wire, 8" OD, full height, No (YO-59940-02).
More Details
Span Gauges
Buy Span Gauges Items Including:
SPAN Pressure Gauge, Vac to 30 psi (YO-68081-02)
SPAN Pressure Gauge, 0 to 100 psi (YO-68081-10)
SPAN Pressure Gauge, 0 to 160 psi (YO-68081-12)
SPAN Pressure Gauge, 0 to 1000 psi (YO-68081-22)
SPAN Pressure Gauge, 0 to 2000 psi (YO-68081-24).
More Details
High Pressure Hydraulic Pumps
Buy High Pressure Hydraulic Pumps Items Including:
High-pressure piston pump, single-piston, 0.1 to 3.0 mL/min, 2500 psi, (A-60S)
High-pressure piston pump, single-piston, 0.2 to 8.0 mL/min, 5000 psi, (B-100-S)
High-pressure piston pump, single-piston, 0.5 to 5.0 mL/min, 1000 psi, (1384).
More Details
Cole-Parmer Pumps - Metering Accessories
Buy Cole-Parmer Pumps - Metering Accessories Items Including:
Cole-Parmer Syringe Pump Accessories, Continuous Low Pressure PPS Pinc
Cole-Parmer Syringe Pump Accessories, Autofill Medium Pressure PTFE Pi
Cole-parmer Stainless Steel Syringe, 2.5 mL, Viton O-ring, 1/16" Swage.
More Details
Ptfe Shop by Industry
Buy Ptfe Shop by Industry Items Including:
Fluke Calibration (formerly Hart Scientific) Bare Thermistor Probe , P (2740014)
Fluke Calibration (formerly Hart Scientific) Bare Thermistor Probe , P (3376880)
Fluke Calibration (formerly Hart Scientific) Bare Thermistor Probe , P (2740023).
More Details
Pcr Mix
Buy Pcr Mix Items Including:
Cole-Parmer Standard Benchtop Chilling/Heating Block, 100-230 VAC
Cole-Parmer Orbital Chilling/Heating Block, 100 to 230 VAC
Cole-Parmer Two-Position Chilling/Heating Block, 100 VAC
Cole-Parmer Aluminum Block, for 40 x 0.5-mL Microcentrifuge Tubes.
More Details
12v Output
Buy 12v Output Items Including:
Instek SPS-1230 DC Switching Single Output Switching Power Supply, 12V.
More Details
Rs485 Cable
Buy Rs485 Cable Items Including:
2 Meter Communication Cable (RS485) for Pump to Pump use with Cole-Par (788304) (YO-74903-73)
5 Meter Communication Cable (RS485) for Pump to Pump use with Cole-Par (788305) (YO-74903-74).
More Details
Nist Calibration Services
Buy Nist Calibration Services Items Including:
Calibration Service For mass 1 Kg To 25 Kg; Astm 4-7; NIST F; Oiml M1 (YO-17108-34)
Calibration Service For mass Set With Maximum Value Between 0 To 0.5 (YO-17109-70)
Innocal NIST Calibration for Viscometers
Innocal NIST Calibration for Incubators.
More Details
Nunc Microplate
Buy Nunc Microplate Items Including:
Thermo Scientific Nunc™ 96-Well Microplates, Immunology, PS, Standa (YO-01928-22)
Thermo Scientific Nunc™ 96-Well Microplates, Immunology, PS, Standa (YO-01928-32)
Thermo Scientific Nunc 96-Well Microplates, Nontreated, PP, Conical pl (YO-01929-30).
More Details
Cole-Parmer Siphon Tubes
Buy Cole-Parmer Siphon Tubes Items Including:
Siphon tube for 75L (20 gal) tank, 16mm ID x 25 mm OD x 92 cm L (YO-50301-91)
Siphon tube for 115L (30 gal) tank, 16mm ID x 25 mm OD x 92 cm L (YO-50301-92)
Siphon tube for 208 L (55 gal) tank, 16mm ID x 25 mm OD x 92 cm L (YO-50301-93).
More Details
Pe Filter
Buy Pe Filter Items Including:
Thermo Scientific Polyethersulfone (PES) Filter Units; membrane; gride
Thermo Scientific Polyethersulfone (PES) Filter Units; 150 mL, 0.20 m
Thermo Scientific Polyethersulfone (PES) Filter Units; capacity, 115mL
Replacement PE filter for 79700-60.
More Details
Delmhorst J-4
Buy Delmhorst J-4 Items Including:
Delmhorst J-4 Wood Moisture Analog Meter 6 to 30%
Replacement pins - 3/4" pins
Replacement PINS - 4IN.
More Details
Nalgene Products
Buy Nalgene Products Items Including:
Thermo Scientific Nalgene Replacement polypropylene cap, 24 mm
Thermo Scientific Nalgene Replacement polypropylene cap, 28 mm
Thermo Scientific Nalgene Replacement polypropylene cap, 53B
Thermo Scientific Nalgene Replacement polypropylene cap, 83B.
More Details
Bel-Art Products Dry Keeper Desiccator Cabinets
Buy Bel-Art Products Dry Keeper Desiccator Cabinets Items Including:
Dry-Keeper™ Desiccator Cabinets, Horizontal; Non-Electric (H420580001)
Dry-Keeper™ Desiccator Cabinets, Vertical; Non-Electric (H420560001)
Dry-Keeper™ Desiccator Cabinets, Horizontal; Electronic (H420580003).
More Details
Dispensette Bottletop Dispensers
Buy Dispensette Bottletop Dispensers Items Including:
BrandTech DispensetteOrganic Bottletop Dispenser, Digital Volume Setti (4731331)
BrandTech Dispensette Organic Bottletop Dispenser, Digital Volume Sett (4731341)
BrandTech Dispensette Organic Bottletop Dispenser, Digital Volume Sett (4731351).
More Details
Extech Humidity Loggers and Recorders
Buy Extech Humidity Loggers and Recorders Items Including:
Extech SD800 Temperature/Humidity/CO2 Data Logger
Extech Temperature and Humidity USB Data logger (RHT10)
Extech Temperature, Humidity and Barometric Pressure Data Logger (RHT5
Extech RH520A Temperature/Humidity Paperless Recorder with LCD and Det.
More Details
Speed Mixers
Buy Speed Mixers Items Including:
Light-Duty, Dual-Shaft Variable Speed Mixer, 120 VAC (YO-04330-00)
Medium-Duty, Dual-Shaft Variable-Speed Mixer, 120 VAC (YO-04340-00)
Heavy-Duty, Variable Speed Mixer, 120 VAC (YO-04340-10)
High-torque, constant-speed mixer, 115 VAC (YO-50402-20).
More Details
Extech Ea31
Buy Extech Ea31 Items Including:
Extech EasyView ™ Light Meter with NIST Certificate.
More Details
Nor-Lake Laboratory Equipment
Buy Nor-Lake Laboratory Equipment Items Including:
Nor-Lake Scientific Select™ Pass-thru Refrigerator +4C, Four door 2
Nor-Lake Scientific Premier Refrigerator, Three Door 80 Cubic Ft
115V
Nor-Lake Scientific Premier Refrigerator, Three Glass Door 80 Cubic Ft.
More Details
Cole-Parmer Flow Rate/Totalizers and Batch Controllers
Buy Cole-Parmer Flow Rate/Totalizers and Batch Controllers Items Including:
Flow rate/totalizer, pulse input, two relays, analog output (57751403)
Flow rate/totalizer, analog input (57751420)
Flow rate/totalizer, analog input, two relays, analog output (57751423)
Batch controller, pulse input, two relays (57751411).
More Details
Glass Refrigerator Doors
Buy Glass Refrigerator Doors Items Including:
Refrigerator, Blood Bank 52 cu ft with Casters, 2 Glass Doors, 115V/60 (NSBR522WWG/0)
Refrigerator, Blood Bank 52 cu ft with Casters, 2 Glass Doors, 230V/50 (NSBR522WWG/5).
More Details
Cole-Parmer Rubber-Top Turntables
Buy Cole-Parmer Rubber-Top Turntables Items Including:
Turntable, 10" Dia.
More Details
Zymit Cleaning Solutions
Buy Zymit Cleaning Solutions Items Including:
Cole-Parmer Zymit™ enzyme cleaner, 1 liter bottle, case of twelve.
More Details
Ansell Cut-Resistant Gloves
Buy Ansell Cut-Resistant Gloves Items Including:
Ansell® HyFlex®Cut Resistant Dyneema® Work Gloves, Size 8 (288736)
Ansell® HyFlex®Cut Resistant Dyneema® Work Gloves, Size 9 (288737)
Ansell® HyFlex®Cut Resistant Dyneema® Work Gloves, Size 10 (288738).
More Details
Cole-Parmer Ferrules
Buy Cole-Parmer Ferrules Items Including:
Graphite straight ferrules, 1/8"; 10/pack (YO-01992-01)
Graphite straight ferrules, 1/4"; 10/pack (YO-01992-02)
Ferrule and nut assembly, stainless steel, 1/16" (YO-31212-67)
Ferrule and nut assembly, brass, 1/16" (YO-31212-68).
More Details
Lamotte Dissolved Oxygen Test Kits
Buy Lamotte Dissolved Oxygen Test Kits Items Including:
LaMotte Dissolved Oxygen Test Kit Refill; Winkler Titration Method, 50.
More Details
Colder all-Plastic Quick-Disconnect Couplings (Apc Series)
Buy Colder all-Plastic Quick-Disconnect Couplings (Apc Series) Items Including:
Quick-disconnect fittings, Polycarbonate fittings, PP, 3/8" ID, insert (MPC22006T03M)
Quick-disconnect fittings, Polysulfone fittings, 1/4" ID, insert, 5/pa (MPC22004T39M)
Coupling Body, Valved, 10mm OD Tube, 10/pack (APCD130M10SH).
More Details
Refill Kit
Buy Refill Kit Items Including:
Refill kit, Phenol test, for 00290-30
Refill kit, Copper, total soluble test, for 00290-50
Refill kit, Phosphate test, for 00290-52
Refill kit, Iron, total soluble test, for 00291-40
Refill kit for Gas Purifier, item number 01418-50 (27069 RF 68).
More Details
70618
results
<<<Previous
5
6
7
8
9
10
11
12
13
Next >>>
View by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
Page:
1
2
3
4
5
6
7
8
9
10
11
12
13
14
15
16
17
18
19
20
21
22
23
24
25
26
27
28
29
30
40
50
60
70
80
90
100
200
300
400
500
600
700
800
900
1,000
2,000
2,354
Tubing Aluminum to Refill Kit
>
31125329-s-toc_2 31125329stoc2